Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Nutrition Research > Learning Center > Plant Profiler > Garlic (Allium sativum)
Plant Profiler
Garlic (Allium sativum)
<em>Allium sativum</em> Image
Synonyms / Common Names / Related Terms
2-propenesulfenic acid, aged garlic extract, aglio, ail, ail commun, ajo, ajoene, akashneem, alisat, allicin, Allii sativi bulbus, alliinase, allium, allyl mercaptan, alubosa elewe, Amaryllidaceae (family), ayo-ishi, ayu, banlasun, camphor of the poor, clove garlic, da-suan, dai toan, dasuan, dawang, diallyl disulphide, diallyl sulfide, diallyl sulphide, dipropyl disulphide, dipropyl sulphide, dra thiam, foom, garlic clove, garlic corns, garlic extract, garlic oil, Gartenlauch, hom khaao, hom kia, hom thiam, hua thiam, kesumphin, kitunguu-sumu, knoblauch, kra thiam, Krathiam, krathiam cheen, krathiam khaao, Kwai®, Kyolic, l'ail, lahsun, lai, la-juan, lasan, lashun, la-suan, lasun, lasuna, lauch, lay, layi, lehsun, lesun, Liliaceae (family), lobha, majo, naharu, nectar of the gods, Ninniku, pa-se-waa, poor man's treacle, rason, rasonam, rasun, rust treacle, rustic treacles, S-allylcysteine (SAC), seer, skordo, sluon, stinking rose, sudulunu, tafanuwa, ta-suam, ta-suan, tellagada, Tellagaddalu, thiam, Thioallyl derivative, Thiosulfinates, toi thum, tum, umbi bawang putih, vallaippundu, Velluli, vellulli, verum, Vinyl dithiin.
Mechanism of Action
Pharmacology:
  • Constituents: Chemical analysis in the 1800s attributed garlic's activity to the sulfur containing garlic oil. In the mid 1900s an American chemist named the strong smelling liquid "allicin".18 The sulfur compound alliin (S-allyl-l-cysteine sulfoxide) produces allicin (diallyl thiosulfinate) via the enzyme allinase when the bulb is crushed or ground. Other sulfur compounds, peptides, steroids, terpenoids, flavonoids, and phenols have increasingly been identified as possible active ingredients19,20,10 as allicin is metabolized. The exact mechanism of action underlying garlic's effects remains unknown and may vary according to the preparation21 and the therapeutic effect.
  • Garlic appears to exert numerous effects on the cardiovascular system, and Atherosclerosis in particular, beyond the reduction of serum lipids. There are possibly multiple protective effects of garlic22,12,23, including inhibition of platelet aggregation and enhancement of fibrinolysis. Wild garlic (Allium ursinum) has been reported to contain similar amounts of sulfur-containing compounds (thiosulfinates and ajoenes) as garlic (Allium sativum), and to exert similar effects on cyclooxygenase, 5-lipoxygenase, angiotensin converting enzyme, and platelet aggregation.24
  • Lipid-lowering Effects: Garlic's lipid lowering effects may occur via inhibition of HMG-CoA reductase or other enzymes25,26,27,28,29,30,31,32, possibly by diallyl di- and trisulphide components of garlic.33,34 Other suggested mechanisms include increased loss of bile salts in feces and mobilization of tissue lipids into circulation35, as garlic has a profound effect on post-prandial hyperlipidemia.36 Wild garlic (Allium ursinum) has shown similar efficacy to garlic (Allium sativum) in decreasing hepatocyte cholesterol synthesis In vitro.24,37 Aged garlic extract and its constituents have been shown to inhibit Cu2+-induced oxidative modification of low-density lipoprotein.38 Aged garlic extract and its constituent S-allylcysteine have been found to protect vascular endothelial cells from injury caused by oxidized LDL.39
  • Animal and human cell lines have demonstrated reductions in vascular tissue lipids40,41, fatty streak formation and atherosclerotic plaque size.22,42,43,44,12,40,45 The mechanism of action may include reduction of lipoprotein oxidation, as demonstrated In vitro38,39,46 and in vivo47, possibly due to organosulfur compounds in garlic.48 However, this hypothesis has been in dispute based on a six-month trial in moderately hypercholesterolemic volunteers which failed to demonstrate any effects of garlic supplementation on lipoprotein oxidation.49
  • Platelet Effects: Garlic and its derived compound ajoene have demonstrated inhibition of platelet aggregation In vitro and in animals50,51,52,15,9,53,54,55,56,57,58,59, and reduction of platelet-dependent thrombus formation.60,61 Research has demonstrated inhibition of platelet aggregation in hypercholesterolemic men62, in healthy subjects, in patients with coronary artery disease63,64,65,66,67, and in subjects with cerebrovascular risk factors3,11. However, a study using a low dietary dosage found no such effects.68 Raw garlic has been shown to inhibit platelet cyclooxygenase In vitro.50 Dose-dependent inhibition of cyclooxygenase in human placenta villi was observed with garlic and with allicin-negative (acid washed) garlic.69 Anti-platelet activity may be attributable to garlic constituents including adenosine, allicin and paraffinic polysulfides.70 Compared to raw garlic, a boiled aqueous garlic extract demonstrated an approximately 50% decrease in platelet aggregation at identical concentrations50, 59, suggesting that cooking garlic may reduce anti-platelet effects. Raw garlic has been shown to reduce serum thromboxane B2 in animal and human research63, 35, at a dose of one clove per day71. However, boiling garlic prior to administration appears to reduce or abolish this effect64, again suggesting a negative impact of cooking on garlic's anti-platelet activity.
  • Fibrinolytic Effects: Increased fibrinolytic activity may account for some degree of garlic's anti-clotting effects72, 20, 73, involving fibrinogen and plasminogen16. Both raw garlic and fried garlic have demonstrated significant increases in fibrinolytic activity in man74, as well as essential oil from raw garlic36. An increase in fibrinolytic activity in patients with ischemic heart disease was found to be maintained after 7-8 weeks of continued therapy.75, 76 However, one study reported that fibrinolytic activity returned to pre-treatment levels after 12 weeks of continuous garlic therapy.76
  • Vascular Effects: Vasorelaxant properties of garlic have been noted in multiple pre-clinical studies.77, 78, 79 Cutaneous microperfusion is increased in humans following ingestion of 600mg of garlic80, 81, 82, and vasodilation of conjunctival arterioles and venules occurs at 900mg.83 Garlic may act on the nitric oxide system84, 85, 86 and exert effects on the elastic properties of vasculature87, yielding changes in systemic blood pressure86. It has been suggested that allicin is the component of garlic responsible for nitric oxide-mediated effects.88 Prostaglandins have been identified in garlic extracts which may exert pharmacologic effects89, although this has not been demonstrated in vivo.
  • Chemoprotective/Anti-tumor Effects: Animal studies have reported protective effects of garlic against hepatotoxins6, 90, 7, 8, cyclophosphamide91, adriamycin92 methylcholanthrene93, gentamicin5, 4-nitroquinoline 1-oxide94, and bromobenzene95, 96. Garlic has demonstrated strong inhibition of cancer development in the presence of known tumor promoters including 12-O-tetradecanoylpharbol-13-acetate97, 98 and 7,12-dimethylbenzanthracene99, and phorbol-myristate-acetate100, as well as tumor inducers such as 7,12-dimethylbenzanthracene98 and 1,2-dimethylhydrazine101,102. There is some evidence that the chemical constituents containing allyl groups may be responsible for chemoprotective properties.103 Research has provided evidence of anti-proliferative effects of garlic on human cancer cell lines104, including induction of apoptosis105,106,107,108,109, regulation of cell cycle progression110, and signal transduction modification. Both cellular proliferation111,112 and immune function appear to be affected113,114.
  • Immunologic Effects: The immunologic activity of garlic may include enhanced phagocytosis, lymphocyte proliferation, enhanced killer cell activity and cytokine production, and prevention of immune suppression.115,116,117,118,119,120 There has been suggestion that heating may adversely affect this benefit, related to a loss of alliinase activity121, and that different preparations have variant pharmacologic activities21.
  • Antimicrobial Effects (Bacteria, Fungi, Yeast): Garlic has been demonstrated In vitro to exert activity against multiple pathogens, including bacteria122,123,14,124,125,10,126,127, including resistant strains128, mycobacteria129,130,131, Helicobacter pylori132,133,10, and fungi134,135. Garlic extract has been found to be bactericidal to Histoplasma capsulatum.136 Ajoene alone possesses antibacterial activity against both gram positive and negative bacterial species and inhibits yeast growth In vitro, and the disulfide bond in ajoene may be responsible for these effects.126 The substrate alliin and its enzyme are found in separate but adjacent compartments of the garlic clove, and appear to exert antimicrobial activity when they are joined. Allicin may act via inhibition of thiol-containing and other enzyme systems, DNA, RNA and protein synthesis.124,137,138,20 It has been suggested that garlic oil's antimicrobial activity is more potent than garlic powder on a unit weight basis.139
  • The minimum inhibitory concentration (MIC) of aqueous garlic extract (AGE) against six clinical yeast isolates ranged between 0.8-1.6mg/mL in one study.138 Garlic appeared to alter the structure and integrity of the outer surface of yeast cells as well as decrease their total lipid content. Garlic was also shown to increase phosphatidylserines while decreasing phosphatidylcholines. Oxygen consumption of yeast cells was also reduced by garlic. The anti-candidal activity of AGE was antagonized by thiols including L-cysteine, glutathione and 2-mercaptoethanol. The effect of AGE on the macromolecular synthesis of Candida albicans revealed protein and nucleic acid syntheses to be inhibited, and lipid synthesis to be arrested.140 Antagonism of lipid synthesis may be a component of the anti-candidal activity of garlic.
  • Anti-viral Effects: In vitro studies have demonstrated effects against several viruses141, including influenza B virus, herpes simplex virus type 1141, herpes simplex virus type 2, parainfluenza virus type 3, vaccinia virus, vesicular stomatitis virus, human rhinovirus type 21,2, and cytomegalovirus4. Weber reported that the compound ajoene, found in oil-macerates of garlic, possesses a high level of antiviral activity followed by allicin, allyl methyl thiosulfinate and methyl allyl thiosulfinate.2
  • Glycemic Effects: SACS (S-allyl cysteine sulfoxide), an antioxidant isolated from garlic, has been found to significantly stimulate insulin secretion from beta cells isolated from normal rats.13 However, multiple human studies have failed to demonstrate glycemic effects of garlic.
  • Genitourinary Effects: Chronic garlic ingestion for 70 days has been associated with suppression of spermatogenesis in rats.17
Pharmacodynamics/Kinetics:
  • Garlic compounds are rapidly absorbed through mucous membranes and skin. Excretion is primarily via liver, kidney and intestines.20
  • Nagae142 described the pharmacokinetics of S-Allylcysteine (SAC) in animal models. The authors demonstrated a first pass effect following rapid gastrointestinal absorption; with liver and kidney metabolism.
  • The pharmacokinetics of the vinyldithiins, transformation products of allicin, have been described; maximal concentrations 15-30 minutes after oral absorption.143
References
  1. Hughes BG, Murray BK, North JA, et al. Antiviral constituents from Allium sativum. Planta Med 1989;55:114.
  2. Weber ND, Andersen DO, North JA, et al. In vitro virucidal effects of Allium sativum (garlic) extract and compounds. Planta Med 1992;58(5):417-423. 1470664
  3. Kiesewetter H, Jung F, Jung EM, et al. Effects of garlic coated tablets in peripheral arterial occlusive disease. Clin Investig 1993;71(5):383-386. 8508009
  4. Guo NL, Lu DP, Woods GL, et al. Demonstration of the anti-viral activity of garlic extract against human cytomegalovirus In vitro. Chin Med J (Engl) 1993;106(2):93-96. 8389276
  5. Pedraza-Chaverri J, Maldonado PD, Medina-Campos ON, et al. Garlic ameliorates gentamicin nephrotoxicity: relation to antioxidant enzymes. Free Radic Biol Med 2000;29(7):602-611. 11033412
  6. Hikino H, Tohkin M, Kiso Y, et al. Antihepatotoxic actions of Allium sativum bulbs. Planta Med 1986;(3):163-168. 3749336
  7. Wang EJ, Li Y, Lin M, et al. Protective effects of garlic and related organosulfur compounds on acetaminophen-induced hepatotoxicity in mice. Toxicol Appl Pharmacol 1996;136(1):146-154. 8560468
  8. Rahmy TR, Hemmaid KZ. Prophylactic action of garlic on the histological and histochemical patterns of hepatic and gastric tissues in rats injected with a snake venom. J Nat Toxins 2001;10(2):137-165. 11405277
  9. Lawson LD, Ransom DK, Hughes BG, et al. Inhibition of whole blood platelet-aggregation by compounds in garlic clove extracts and commercial garlic products. Thrombosis Res 1992;65(2):141-156.
  10. Sivam GP. Protection against Helicobacter pylori and other bacterial infections by garlic. J Nutr 2001;131(3s):1106S-1108S. 11238826
  11. Czerny B, Samochowiec J. Klinische Untersuchungen mit einem Knoblauch-Lezithin-Präparat. Arztezeitschr Naturheilverf 1996;37:126-129.
  12. Koscielny J, Klussendorf D, Latza R, et al. The antiatherosclerotic effect of Allium sativum. Atherosclerosis 1999;144:237-249.
  13. Augusti KT, Sheela CG. Antiperoxide effect of S-allyl cysteine sulfoxide, an insulin secretagogue, in diabetic rats. Experientia 1996;52(2):115-120. 8608811
  14. Kumar M, Berwal JS. Sensitivity of food pathogens to garlic (Allium sativum). J Appl Microbiol 1998;84(2):213-215. 9633635
  15. Apitz-Castro R, Escalante J, Vargas R, et al. Ajoene, the antiplatelet principle of garlic, synergistically potentiates the antiaggregatory action of prostacyclin, forskolin, indomethacin and dypiridamole on human platelets. Thromb Res 1986;42(3):303-311. 3520940
  16. Harenberg J, Giese C, Zimmermann R. Effect of dried garlic on blood coagulation, fibrinolysis, platelet aggregation and serum cholesterol levels in patients with hyperlipoproteinemia. Atherosclerosis 1988;74(3):247-249. 3240334
  17. Dixit VP, Joshi S. Effects of chronic administration of garlic (Allium sativum Linn) on testicular function. Indian J Exp Biol 1982;20(7):534-536. 7173985
  18. Block E. The chemistry of garlic and onions. Sci Am 1985;252(3):114-119. 3975593
  19. Agarwal K. Therapeutic actions of garlic constituents. Medicinal Res Rev 1996;16(1):111-124.
  20. Reuter HD, Sendl A. Allium sativum and Allium ursinum: chemistry, pharmacology and medicinal applications. In: Wagner H, Farnsworth NR, editors. Economic and Medicinal Research. London: Academic Press Ltd, 1994: 55-103.
  21. Kasuga S, Uda N, Kyo E, et al. Pharmacologic activities of aged garlic extract in comparison with other garlic preparations. J Nutr 2001;131(3s):1080S-1084S. 11238821
  22. Campbell JH, Efendy JL, Smith NJ, et al. Molecular basis by which garlic suppresses Atherosclerosis. J Nutr 2001;131(3s):1006S-1009S. 11238806
  23. Patumraj S, Tewit S, Amatyakul S, et al. Comparative effects of garlic and aspirin on diabetic cardiovascular complications. Drug Deliv 2000;7(2):91-96. 10892409
  24. Sendl A, Elbl G, Steinke B, et al. Comparative pharmacological investigations of Allium ursinum and Allium sativum. Planta Med 1992;58(1):1-7. 1620734
  25. Chi MS. Effects of garlic products on lipid metabolism in cholesterol-fed rats (41494). Proc Soc Exper Biol Med 1982;171:174-178.
  26. Chi M, Koh ET, Stewart TJ. Effects of garlic on lipid metabolism in rats fed cholesterol or lard. J Nutrit 1982;112(2):241-248.
  27. Qureshi A, Abuirmeileh N, Din Z, et al. Inhibition of cholesterol and fatty acid biosynthesis in liver enzymes and chicken hepatocytes by polar fractions of garlic. Lipids 1983;18:343-348.
  28. Qureshi AA, Crenshaw TD, Abuirmeileh N. Influence of minor plant constituents on porcine hepatic lipid metabolism. Atherosclerosis 1987;64:109-115.
  29. Gebhardt R, Beck H, Wagner K. Inhibition of cholesterol biosynthesis by allicin and ajoene in rat hepatocytes and HepG2 cells. Biochim Biophys Acta 1994;1213(1):57-62.
  30. Yeh YY, Yeh SM. Garlic reduces plasma lipids by inhibiting hepatic cholesterol and triacylglycerol synthesis. Lipids 1994;29(3):189-193. 8170288
  31. Gupta N, Porter TD. Garlic and garlic-derived compounds inhibit human squalene monooxygenase. J Nutr 2001;131(6):1662-1667. 11385050
  32. Yeh YY, Liu L. Cholesterol-lowering effect of garlic extracts and organosulfur compounds: human and animal studies. J Nutr 2001;131(3s):989S-993S. 11238803
  33. Gebhardt R, Beck H. Differential inhibitory effects of garlic-derived organosulfur compounds on cholesterol biosynthesis in primary rat hepatocyte cultures. Lipids 1996;31(12):1269-1276.
  34. Liu L, Yeh YY. Inhibition of cholesterol biosynthesis by organosulfur compounds derived from garlic. Lipids 2000;35(2):197-203. 10757551
  35. Bordia A, Verma SK, Srivastava KC. Effect of garlic (Allium sativum) on blood lipids, blood sugar, fibrinogen and fibrinolytic activity in patients with coronary artery disease. Prostaglandins Leukot Essent Fatty Acids 1998;58(4):257-263. 9654398
  36. Bordia A, Bansal HC. Letter: Essential oil of garlic in prevention of Atherosclerosis. Lancet 1973;2(7844):1491-1492. 4129327
  37. Sendl A, Schliack M, Loser R, et al. Inhibition of cholesterol synthesis In vitro by extracts and isolated compounds prepared from garlic and wild garlic. Atherosclerosis 1992;94(1):79-85. 1632861
  38. Ide N, Nelson AB, Lau BHS. Aged garlic extract and its constituents inhibit Cu2+ -induced oxidative modification of low density lipoprotein. Planta Med 1997;63:263-264.
  39. Ide N, Lau BH. Garlic compounds protect vascular endothelial cells from oxidized low density lipoprotein-induced injury. J Pharm Pharmacol 1997;49(9):908-911. 9306260
  40. Orekhov A, Tertov V. In vitro effect of garlic powder extract on lipid content in normal and atherosclerotic human aortic cells. Lipids 1997;32:1055-1060.
  41. Orekhov AN, Tertov VV, Sobenin IA, et al. Direct anti-Atherosclerosis-related effects of garlic. Ann Med 1995;27(1):63-65. 7742001
  42. Efendy JL, Simmons DL, Campbell GR, et al. The effect of the aged garlic extract, 'Kyolic', on the development of experimental Atherosclerosis. Atherosclerosis 1997;132(1):37-42. 9247357
  43. Jain RC, Konar DB. Effect of garlic oil in experimental cholesterol Atherosclerosis. Atherosclerosis 1978;29(2):125-129. 646845
  44. Kiesewetter H. Long-term effect of garlic powder tablets on the development of plaque formation in the carotid branches of both femoral arteries - a preliminary report. Eur J Clin Res 1996;8:34-35.
  45. Durak Ì, Öztürk HS, Olcay E, et al. Effects of garlic extract supplementation on blood lipid and antioxidant parameters and atherosclerotic plaque formation process in cholesterol-fed rabbits. J Herbal Pharmacother 2002;2(2):19-32.
  46. Munday JS, James KA, Fray LM, et al. Daily supplementation with aged garlic extract, but not raw garlic, protects low density lipoprotein against In vitro oxidation. Atherosclerosis 1999;143(2):399-404.10217370
  47. Phelps S, Harris WS. Garlic supplementation and lipoprotein oxidation susceptibility. Lipids 1993;28(5):475-477. 8316057
  48. Dwivedi C, John LM, Schmidt DS, et al. Effects of oil-soluble organosulfur compounds from garlic on doxorubicin-induced lipid peroxidation. Anticancer Drugs 1998;9(3):291-294. 9625441
  49. Byrne DJ, Neil HA, Vallance DT, et al. A pilot study of garlic consumption shows no significant effect on markers of oxidation or sub-fraction composition of low-density lipoprotein including lipoprotein(a) after allowance for non-compliance and the placebo effect. Clin Chim Acta 1999;285(1-2):21-33. 10481920
  50. Ali M. Mechanism by which garlic (Allium sativum) inhibits cyclooxygenase activity. Effect of raw versus boiled garlic extract on the synthesis of prostanoids. Prostaglandins Leukot Essent Fatty Acids 1995;53(6):397-400. 8821119
  51. Teranishi K, Apitz-Castro R, Robson SC, et al. Inhibition of baboon platelet aggregation In vitro and in vivo by the garlic derivative, ajoene. Xenotransplantation 2003;10(4):374-379. 12795687
  52. Apitz-Castro R, Cabrera S, Cruz MR, et al. Effects of garlic extract and of three pure components isolated from it on human platelet aggregation, arachidonate metabolism, release reaction and platelet ultrastructure. Thromb Res 1983;32(2):155-169. 6419374
  53. Makheja AN, Vanderhoek JY, Bailey JM. Inhibition of platelet aggregation and thromboxane synthesis by onion and garlic. Lancet 1979;1(8119):781. 86023
  54. Srivastava KC, Tyagi OD. Effects of a garlic-derived principle (ajoene) on aggregation and arachidonic acid metabolism in human blood platelets. Prostaglandins Leukot Essent Fatty Acids 1993;49(2):587-595. 8415808
  55. Boullin DJ. Garlic as a platelet inhibitor. Lancet 1981;1(8223):776-777. 6110972
  56. Jamaluddin MP, Krishnan LK, Thomas A. Ajoene inhibition of platelet aggregation: possible mediation by a hemoprotein. Biochem Biophys Res Communicat 1988;153(1):479-486.
  57. Rendu F, Daveloose D, Debouzy JC, et al. Ajoene, the antiplatelet compound derived from garlic, specifically inhibits platelet release reaction by affecting the plasma membrane internal microviscosity. Biochem Pharmacol 1989;38(8):1321-1328. 2539823
  58. Oshiba S, Sawai H, Tamada T, et al. [Inhibitory effect of orally administered inclusion complex of garlic oil on platelet aggregation in man]. Igaku no Ayuma 1990;155(3):199-200.
  59. Ali M, Bordia T, Mustafa T. Effect of raw versus boiled aqueous extract of garlic and onion on platelet aggregation. Prostaglandins Leukot Essent Fatty Acids 1999;60(1):43-47. Abstract
  60. Apitz-Castro R, Badimon JJ, Badimon L. Effect of ajoene, the major antiplatelet compound from garlic, on platelet thrombus formation. Thromb Res 1992;68(2):145-155. 1475777
  61. Apitz-Castro R, Badimon JJ, Badimon L. A garlic derivative, ajoene, inhibits platelet deposition on severely damaged vessel wall in an in vivo porcine experimental model. Thromb Res 1994;75(3):243-249. 7992235
  62. Steiner M, Lin RS. Changes in platelet function and susceptibility of lipoproteins to oxidation associated with administration of aged garlic extract. J Cardiovasc Pharmacol 1998;31(6):904-908. 9641475
  63. Bordia A, Verma SK, Srivastava KC. Effect of garlic on platelet aggregation in humans: a study in healthy subjects and patients with coronary artery disease. Prostaglandins Leukot Essent Fatty Acids 1996;55(3):201-205. 8931120
  64. Bordia T, Mohammed N, Thomson M, et al. An evaluation of garlic and onion as antithrombotic agents. Prostaglandins Leukot Essent Fatty Acids 1996;54(3):183-186. 8860105
  65. Legnani C, Frascaro M, Guazzaloca G, et al. Effects of a dried garlic preparation on fibrinolysis and platelet aggregation in healthy subjects. Arzneimittelforschung 1993;43(2):119-122. 8457236
  66. Rahman K, Billington D. Dietary supplementation with aged garlic extract inhibits ADP-induced platelet aggregation in humans. J Nutr 2000;130(11):2662-2665. 11053504
  67. Steiner M, Li W. Aged garlic extract, a modulator of cardiovascular risk factors: a dose- finding study on the effects of AGE on platelet functions. J Nutr 2001;131(3s):980S-984S. 11238801
  68. Morris J, Burke V, Mori TA, et al. Effects of garlic extract on platelet aggregation: a randomized placebo- controlled double-blind study. Clin Exp Pharmacol Physiol 1995;22(6-7):414-417. 8582091
  69. Das I, Patel S, Sooranna SR. Effects of aspirin and garlic on cyclooxygenase-induced chemiluminescence in human term placenta. Biochem Soc Transl 1997;25(1):99S. 9056997
  70. Makheja AN, Bailey JM. Antiplatelet constituents of garlic and onion. Agents Actions 1990;29(3-4):360-363. 2111084
  71. Ali M, Thomson M. Consumption of a garlic clove a day could be beneficial in preventing thrombosis. Prostaglandins Leukot Essent Fatty Acids 1995;53(3):211-212. 7480084
  72. Kendler BS. Garlic (Allium sativum) and onion (Allium cepa): a reof their relationship to cardiovascular disease. Prev Med 1987;16(5):670-685. 3317392
  73. Srivastava KC, Bordia A, Verma SK. Garlic (Allium sativum) for disease prevention. S Afr J Sci 1995;91:68-77.
  74. Chutani SK, Bordia A. The effect of fried versus raw garlic on fibrinolytic activity in man. Atherosclerosis 1981;38(3-4):417-421. 7225179
  75. Bordia AK, Joshi HK, Sanadhya YK, et al. Effect of essential oil of garlic on serum fibrinolytic activity in patients with coronary artery disease. Atherosclerosis 1977;28(2):155-159. 911374
  76. Arora RC, Arora S, Gupta RK. The long-term use of garlic in ischemic heart disease--an appraisal. Atherosclerosis 1981;40(2):175-179. 7306257
  77. Ashraf MZ, Hussain ME, Fahim M. Endothelium mediated vasorelaxant response of garlic in isolated rat aorta: role of nitric oxide. J Ethnopharmacol 2004;90(1):5-9. 14698500
  78. Baluchnejadmojarad T, Roghani M, Homayounfar H, et al. Beneficial effect of aqueous garlic extract on the vascular reactivity of streptozotocin-diabetic rats. J Ethnopharmacol 2003;85(1):139-144. 12576212
  79. Baluchnejadmojarad T, Roghani M. Endothelium-dependent and -independent effect of aqueous extract of garlic on vascular reactivity on diabetic rats. Fitoterapia 2003;74(7-8):630-637. 14630166
  80. Jung EM, Jung F, Mrowietz C, et al. Influence of garlic powder on cutaneous microcirculation. A randomized placebo-controlled double-blind cross-over study in apparently healthy subjects. Arzneimittelforschung 1991;41(6):626-630. 1930351
  81. Jung F, Jung EM, Mrowietz C, et al. [The effects of garlic powder on cutaneous microcirculation. A cross-over test with healthy test persons]. Med Welt 1991;42:28-30.
  82. Wohlrab J, Wohlrab D, Marsch WC. Acute effect of a dried ethanol-water extract of garlic on the microhaemovascular system of the skin. Arzneimittelforschung 2000;50(7):606-612. 10965416
  83. Wolf S, Reim M, Jung F. Effect of garlic on conjunctival vessels: a randomised, placebo- controlled, double-blind trial. Br J Clin Pract Suppl 1990;69:36-39. 2083172
  84. Das I, Khan NS, Sooranna SR. Potent activation of nitric oxide synthase by garlic: a basis for its therapeutic applications. Curr Med Res Opin 1995;13(5):257-263. 7555034
  85. Dirsch VM, Kiemer AK, Wagner H, et al. Effect of allicin and ajoene, two compounds of garlic, on inducible nitric oxide synthase. Atherosclerosis 1998;139:333-339.
  86. Pedraza-Chaverri J, Tapia E, Medina-Campos O. Garlic prevents hypertension induced by chronic inhibition of nitric oxide synthesis. Life Sciences 1998;62(6):71-77.
  87. Breithaupt-Grogler K, Ling M, Boudoulas H, et al. Protective effect of chronic garlic intake on elastic properties of aorta in the elderly. Circulation 1997;96(8):2649-2655. 9355906
  88. Ku DD, Abdel-Razek TT, Dai J, et al. Mechanisms of garlic induced pulmonary vasorelaxation: role of allicin. Circulation 1997;96(8S):6-I.
  89. Al-Naghdy SA, Abdel-Rahman MO, Heiba HI. Evidence for some prostaglandins in Allium sativum extracts. Phytother Res 1988;2(4):196-197.
  90. Kweon S, Park KA, Choi H. Chemopreventive effect of garlic powder diet in diethylnitrosamine-induced rat hepatocarcinogenesis. Life Sci 2003;73(19):2515-2526. 12954459
  91. Unnikrishnan MC, Soudamini KK, Kuttan R. Chemoprotection of garlic extract toward cyclophosphamide toxicity in mice. Nutr Cancer 1990;13(3):201-207. 2308875
  92. Thabrew MI, Samarawickrema NA, Chandrasena LG, et al. Protection by garlic against adriamycin induced alterations in the oxido-reductive status of mouse red blood cells. Phytother Res 2000;14(3):215-217. 10815020
  93. Hussain SP, Jannu LN, Rao AR. Chemopreventive action of garlic on methylcholanthrene-induced carcinogenesis in the uterine cervix of mice. Cancer Lett 1990;49(2):175-180. 2306712
  94. Balasenthil S, Ramachandran CR, Nagini S. Prevention of 4-nitroquinoline 1-oxide-induced rat tongue carcinogenesis by garlic. Fitoterapia 2001;72(5):524-531. 11429247
  95. Wang BH, Zuzel KA, Rahman K, et al. Protective effects of aged garlic extract against bromobenzene toxicity to precision cut rat liver slices. Toxicology 1998;126(3):213-222. Abstract
  96. Wang BH, Zuzel KA, Rahman K, et al. Treatment with aged garlic extract protects against bromobenzene toxicity to precision cut rat liver slices. Toxicology 1999;132(2-3):215-225. 10433384
  97. Nishino H, Iwashima A, Itakura Y, et al. Antitumor-promoting activity of garlic extracts. Oncology 1989;46(4):277-280. 2500627
  98. Dwivedi C, Rohlfs S, Jarvis D, et al. Chemoprevention of chemically induced skin tumor development by diallyl sulfide and diallyl disulfide. Pharm Res 1992;9(12):1668-1670. 1488416
  99. Balasenthil S, Rao KS, Nagini S. Retinoic acid receptor-beta mRNA expression during chemoprevention of hamster cheek pouch carcinogenesis by garlic. Asia Pac J Clin Nutr 2003;12(2):215-218. 12810414
  100. Belman S. Onion and garlic oils inhibit tumor promotion. Carcinogenesis 1983;4(8):1063-1065. 6872151
  101. Wargovich MJ. Diallyl sulfide, a flavor component of garlic (Allium sativum), inhibits dimethylhydrazine-induced colon cancer. Carcinogenesis 1987;8(3):487-489. 3815744
  102. Wargovich MJ, Uda N, Woods C, et al. Allium vegetables: their role in the prevention of cancer. Biochem Soc Transl 1996;24(3):811-814. 8878853
  103. Sparnins VL, Barany G, Wattenberg LW. Effects of organosulfur compounds from garlic and onions on benzo[a]pyrene-induced neoplasia and glutathione S-transferase activity in the mouse. Carcinogenesis 1988;9(1):131-134. 3335037
  104. Pinto JT, Rivlin RS. Antiproliferative effects of allium derivatives from garlic. J Nutr 2001;131(3s):1058S-1060S. 11238816
  105. Dirsch VM, Gerbes AL, Vollmar AM. Ajoene, a compound of garlic, induces apoptosis in human promyeloleukemic cells, accompanied by generation of reactive oxygen species and activation of nuclear factor kappaB. Mol Pharmacol 1998;53(3):402-407. 9495804
  106. Xiao D, Pinto JT, Soh JW, et al. Induction of apoptosis by the garlic-derived compound S-allylmercaptocysteine (SAMC) is associated with microtubule depolymerization and c-Jun NH(2)-terminal kinase 1 activation. Cancer Res 2003;63(20):6825-6837. 14583480
  107. Bradley R, Endres J, Hockenberry D, et al. Investigation of garlic-induced apoptosis in breast cancer cell lines [poster presentation]. International Scientific Conference on Complementary, Alternative and Integrative Medicine Research, Boston, MA, 2002.
  108. Hu X, Cao BN, Hu G, et al. Attenuation of cell migration and induction of cell death by aged garlic extract in rat sarcoma cells. Int J Mol Med 2002;9(6):641-643. 12011982
  109. Tilli CM, Stavast-Kooy AJ, Vuerstaek JD, et al. The garlic-derived organosulfur component ajoene decreases basal cell carcinoma tumor size by inducing apoptosis. Arch Dermatol Res 2003;295(3):117-123. 12756587
  110. Li M, Ciu JR, Ye Y, et al. Antitumor activity of Z-ajoene, a natural compound purified from garlic: antimitotic and microtubule-interaction properties. Carcinogenesis 2002;23(4):573-579. 11960908
  111. Morioka N, Sze LL, Morton DL, et al. A protein fraction from aged garlic extract enhances cytotoxicity and proliferation of human lymphocytes mediated by interleukin-2 and concanavalin A. Cancer Immunol Immunother 1993;37(5):316-322. 8402735
  112. Siegers CP, Steffen B, Robke A, et al. The effects of garlic preparation against human tumour cell proliferation. Phytomedicine 1999;6(1):7-11.
  113. Milner JA. A historical perspective on garlic and cancer. J Nutr 2001;131(3s):1027S-1031S. 11238810
  114. Lau BH, Tadi PP, Tosk JM. Allium sativum (garlic) and cancer prevention. Nutrit Res 1990;10:937-948.
  115. Lamm DL, Riggs DR. The potential application of Allium sativum (garlic) for the treatment of bladder cancer. Urol Clin North Am 2000;27(1):157-162, xi. 10696254
  116. Hassan ZM, Yaraee R, Zare N, et al. Immunomodulatory affect of R10 fraction of garlic extract on natural killer activity. Int Immunopharmacol 2003;3(10-11):1483-1489. 12946445
  117. Keiss HP, Dirsch VM, Hartung T, et al. Garlic (Allium sativum L.) modulates cytokine expression in lipopolysaccharide-activated human blood thereby inhibiting NF-kappaB activity. J Nutr 2003;133(7):2171-2175. 12840173
  118. Lamm DL, Riggs DR. Enhanced immunocompetence by garlic: role in bladder cancer and other malignancies. J Nutr 2001;131(3s):1067S-1070S. 11238818
  119. Salman H, Bergman M, Bessler H, et al. Effect of a garlic derivative (alliin) on peripheral blood cell immune responses. Int J Immunopharmacol 1999;21(9):589-597. 10501628
  120. Mel'chinskaia EN, Popovtseva ON, Gromnatskii NI. [Immunologic aspects of alisate in diabetes mellitus patients]. Biulleten' eksperimental'noi biologii i meditsiny 1997;124(11):595-597. 9471268
  121. Song K, Milner JA. The influence of heating on the anticancer properties of garlic. J Nutr 2001;131(3s):1054S-1057S. 11238815
  122. Yoshida H, Iwata N, Katsuzaki H, et al. Antimicrobial activity of a compound isolated from an oil-macerated garlic extract. Biosci Biotechnol Biochem 1998;62(5):1014-1017. 9648236
  123. Hughes BG, Lawson LD. Antimicrobial effects of Allium sativum L. (garlic), Allium ampeloprasum L. (Elephant garlic), and Allium cepa L. (Onion), garlic compounds and commercial garlic supplement products. Phytother Res 1991;5:154-158.
  124. Ankri S, Mirelman D. Antimicrobial properties of allicin from garlic. Microbes Infect 1999;1(2):125-129. 10594976
  125. Sasaki J, Kita T, Ishita K, et al. Antibacterial activity of garlic powder against Escherichia coli O-157. J Nutr Sci Vitaminol (Tokyo) 1999;45(6):785-790. 10737231
  126. Naganawa R, Iwata N, Ishikawa K, et al. Inhibition of microbial growth by ajoene, a sulfur-containing compound derived from garlic. Appl Environ Microbiol 1996;62(11):4238-4242. 8900018
  127. Rao R, Rao S, Natarajan S, et al. Inhibition of Mycobacterium tuberculosis by garlic extract. Nature 1946; (3988):441.
  128. Sharma VD, Sethi MS, Kumar A, et al. Antibacterial property of Allium sativum Linn: in vivo & In vitro studies. Indian J Exper Biol 1977;15:466-468.
  129. Abbruzzese MR, Delaha EC, Garagusi VF. Absence of antimycobacterial synergism between garlic extract and antituberculosis drugs. Diagn Microbiol Infect Dis 1987;8(2):79-85. 3123123
  130. Delaha EC, Garagusi VF. Inhibition of mycobacteria by garlic extract (Allium sativum). Antimicrob Agents Chemother 1985;27(4):485-486. 4004189
  131. Deshpande RG, Khan MB, Bhat DA, et al. Inhibition of Mycobacterium avium complex isolates from AIDS patients by garlic (Allium sativum). J Antimicrob Chemother 1993;32(4):623-626. 8288504
  132. Jonkers D, van den BE, van D, I, et al. Antibacterial effect of garlic and omeprazole on Helicobacter pylori. J Antimicrob Chemother 1999;43(6):837-839. 10404325
  133. O'Gara EA, Hill DJ, Maslin DJ. Activities of garlic oil, garlic powder, and their diallyl constituents against Helicobacter pylori. Appl Environ Microbiol 2000;66(5):2269-2273. 10788416
  134. Moore GS, Atkins RD. The fungicidal and fungistatic effects of an aqueous garlic extract on medically important yeast-like fungi. Mycologia 1977;69(2):341-348. 865503
  135. Caporaso N, Smith SM, Eng RH. Antifungal activity in human urine and serum after ingestion of garlic (Allium sativum). Antimicrob Agents Chemother 1983;23(5):700-702. 6870217
  136. Fliermans CB. Inhibition of Histoplasma capsulatum by garlic. Mycopathol Mycol Appl 1973;50(3):227-231. 4729537
  137. Feldberg RS, Chang SC, Kotik AN, et al. In vitro mechanism of inhibition of bacterial cell growth by allicin. Antimicrob Agents Chemother 1988;32(12):1763-1768. 2469386
  138. Ghannoum MA. Studies on the anticandidal mode of action of Allium sativum (garlic). J Gen Microbiol 1988;134 ( Pt 11):2917-2924. 3076177
  139. Ross ZM, O'Gara EA, Hill DJ, et al. Antimicrobial properties of garlic oil against human enteric bacteria: evaluation of methodologies and comparisons with garlic oil sulfides and garlic powder. Appl Environ Microbiol 2001;67(1):475-480. 11133485
  140. Adetumbi M, Javor GT, Lau BH. Allium sativum (garlic) inhibits lipid synthesis by Candida albicans. Antimicrob Agents Chemother 1986;30(3):499-501. 3535667
  141. Tsai Y, Cole LL, Davis LE, et al. Antiviral properties of garlic: In vitro effects on influenza B, herpes simplex and coxsackie viruses. Planta Med 1985;(5):460-461. 3001801
  142. Nagae S, Ushijima M, Hatono S, et al. Pharmacokinetics of the garlic compound S-allylcysteine. Planta Med 1994;60(3):214-217. 8073085
  143. Egen-Schwind C, Eckard R, Jekat FW, et al. Pharmacokinetics of vinyldithiins, transformation products of allicin. Planta Med 1992;58:8-13.
Licensed by Natural Standard Copyright © 2010 by Natural Standard Corporation. All Rights Reserved.
back to Plant Profiler
back to top