|
Plant Profiler
Astragalus (Astragalus membranaceous)
|
|
|
Synonyms / Common Names / Related Terms
Astragalosides I-IV, Astragalus gummifera, Astragalus lentiginosus, Astragalus membranaceus, Astragalus membranaceus japonica, Astragalus mollissimus, Astragalus mongholicus, astragalus polysaccharide, Astragalus propinquus, astragalus saponin, Astragalus trigonus, astragel, baak kei, beg kei, bei qi, buck qi, calycosin, Fabaceae (family), formononetin, gaba-aminobytyric acid, goat's horn, goat's thorn, green dragon, gum dragon, gum tragacanthae, gummi tragacanthae, hoang ky, hog gum, Huang Qi, huang-chi, huangoi, huangqi, hwanggi, isoflavonoids, ji cao, Leguminosae (family), locoweed, membranous milk vetch, milk vetch, milkvetch, Mongolian milk, Mongolian milk vetch, neimeng hhuangqi, ogi, ononin, ougi, Phaca membranacea Fisch., radix astragali, spino santo, Syrian tragacanth, tai shen, tragacanth, trigonosides I-III, wong kei, yellow leader, yellow vetch, Zhongfengnaomitong.
Mechanism of Action
Pharmacology:
- Constituents: Active constituents of astragalus root have been identified and characterized. These components include: polysaccharide cycloartane glycoside fractions (astragalosides I-IV and trigonosides I-III), four major isoflavonoids (formononetin, ononin, calycosin, and its glycoside), saponins, several minor isoflavonoids,
and other biogenic amines.25,26,27,28,29,8,30,31 Astragalus also contains triterpenoids. Triterpenoids and saponins have a structural similarity to steroid hormone precursors.
- Antidiabetic effects: According to animal and human study, saponins, isoflavones and polysaccharides in astragalus can lower blood glucose.24,5,17,32,33,6,7 Based on experimental study, astragaloside IV, a saponin isolated from Astragalus membranaceus, inhibited protein tyrosine phosphorylase and protein kinase C activation, as well as improving F-actin rearrangements, thereby altering insulin sensitivity.30,34 Several investigators have observed increased in vitro cellular uptake of glucose and increased protein synthesis in adipocytes.35,4 Isoflavones in Astragalus membranaceus, including formononectin and biochanin A, were potent activators of both peroxisome proliferator-activated receptors (PPAR) and receptors that are used to restore glycemic balance.33
- Anti inflammatory effects: The anti-inflammatory activity in vivo may be mediated by inhibition of NF-kappaB activation and mRNA expression.36,37
- Antimicrobial activity: Astragalus polysaccharides have been shown to induce endogenous interferon production in animals and humans and to potentiate the actions of interferon in viral infections.38,9,22,2 Mice pretreated with astragalus and exposed to Coxsachie B3, Japanese encephalitis virus, NDV, and Sendai viruses have significantly higher interferon levels and macrophage production compared to animals that are not pretreated with astragalus.39,21 Several investigators have reported that mice with viral myocarditis exhibited improvement in overall outcomes and cardiac function, as well as reduced myocardial lesions, viral load, and viral replication in response to astragalus.40,41,20,10,42 Astragalus polysaccharide fractions have also been reported to stimulate the production of IgA, IgM, IgG, and antigen-specific splenocytes in response to bacterial infections in poultry.14,15,16 In contrast, astragalus exhibited no in vitro cytotoxic or antiviral activity against HIV infection.43,39
- Antineoplastic properties/amelioration of chemotherapy side effects: There is in vitro evidence that astragalus can potentiate the efficacy and reduce the adverse effects of chemotherapy via stimulation of the immune system.18,19,2 Wei et al. reported that astragalus extract significantly increased in vitro cytokine secretion and gene expression in blood mononuclear cells of lung cancer patients.44 Astragalus extracts have been shown to induce LAK cell activity in the lymphocytes of cancer and HIV patients and stimulate the blastogenic response of lymphocytes to mutagens in patients with various cancers.45,46,47,48,49,50 Lau et al. reported that an astragalus-containing herbal combination inhibited tumor growth in vivo while augmenting macrophage and lymphokine-activated killer cell activities.51 Treatment with astragalus-containing herbal remedies also inhibited tumor growth and prolonged survival in rats compared to rats treated with conventional chemotherapy. Other authors have reported the potentiation of interleukin-2 (IL-2)-generated cytotoxicity in animal models45,52 and reduction of mutagenesis53.
- Antioxidant and cellular protective properties: Astragalus constituents, particularly the flavonoids, exert significant cellular antioxidant effects that appear to be protective against cardiovascular, hepatic, pulmonary, and renal pathological changes.54,55 In vitro, astragalus has been reported to protect against free radical-mediated renal tubular damage induced by high-energy shock waves56 and against radiation injury23. In the hearts of healthy animals and animal models of cardiovascular disease, astragalus polysaccharide and flavonoid fractions have been reported to scavenge mitochondrial free radicals, protect against lipid peroxidation, protect mitochondria from isoproterenol-induced injury, prevent diabetes-induced myocardial hypertrophy, and attenuate pathophysiological decrements in cardiac structure and function in congestive heart failure, diabetes, and experimental hypertension.57,58,59,11,60,61 Astragalus polysaccharide and flavonoid extracts have been shown to preserve kidney function in animal models of renal failure and liver functions in response to hepatotoxins.56,62,63,4,64,65,66 Similarly, astragalus appears to protect cells against hypoxic challenges. Astragaloside IV has been reported to protect human endothelial cells and intestinal epithelial cells against reperfusion injury in vitro following hemorrhagic shock.67 Cellular protection from anoxia has also been observed in cultured neurons68 and in mouse brains after transient focal ischemia69. Astragaloside IV has also inhibited platelet aggregation, decreased plasminogen activation, and facilitated intravascular lysis of fibrin clots in humans and in animal disease models.18,70 In a model of ototoxicity, Xuan et al. reported that astragalus protected cells from pathologic injury via enhanced DNA and RNA synthesis.65 In an animal study, Astragalus membranaceus had preventive and therapeutic potential in experimental colitis.1 The anti-inflammatory actions involved antioxidation along with the inhibition of adhesion molecule synthesis in the colonic tissues. In an in vitro study, astragalus and astragalus saponins potently protected endothelium-dependent relaxation against the acute injury from homocysteine through nitric oxide regulatory pathways, in which antioxidation played a key role.31
- Cardiovascular effects: Experimental studies indicate that astragalosides affect cardiovascular function at the cellular, isolated tissue, and whole-body levels in both animals and humans. In isolated working rat hearts, Wang et al. reported that astragalus extract produced positive inotropic actions at higher doses and negative inotropic effects at lower doses.52 Astragalus also produces hypotensive actions via peripheral vasodilation in healthy and hypertensive animals.71,72 Chen et al. reported that astragalus significantly reduced arterial pressure in the rat model of renal hypertension and improved cardiac function in experimental congestive heart failure.73 This effect was confirmed by Ma et al. who reported that astragalus improved left ventricular systolic function and potentiated the cardiotonic, diuretic, and natriuretic renal responses to atrial natriuretic peptide in rats with experimentally induced chronic heart failure.13 There is in vitro evidence that astragalus may also stimulate angiogenesis and revascularization in ischemic myocardium66 and amnion74 and promote vascular endothelial cell proliferation and DNA synthesis74. Astragalus has been suggested to increase sodium pump activity in erythrocytes.75
- Hormonal/reproductive effects: Astragalus has significant effects on various hormonal systems. Astragalosides are reported to significantly increase growth hormone release in rat pituitary cell culture compared to controls.12 Astragalus polysaccharide fractions have also been shown to prevent bone loss in ovariectomized rats.76 In vitro sperm motility and viability have been shown to be significantly increased in healthy rats and men.77 Astragalus has also been reported to inhibit in rats decrements in sperm production and motility caused by cadmium exposure.25 Liu et al. recently reported that astragalus extract significantly increased in vitro sperm motility and viability in 30 infertile men.78
- Immune system effects: Astragalus flavonoids are reported to increase in vivo cell-mediated immunity in guinea pigs79 and mice42. Astragalus appears to enhance cell-mediated immunity by increasing the number of T-helper cell type 2 cytokines, increasing levels of tumor necrosis factor and interleukin-6.80,81,48,82,83,84 It has been shown to stimulate macrophage and natural killer cell activity.45,85,3 According to animal study, astragalus enhanced both cellular and humoral immune responses to bacterial infections when compared to controls.15 Marrow blood collected from patients with ischemia of the lower limbs and exposed to astragalus had a significantly increased ratio of CD34+ cells (p<0.05).86
Pharmacodynamics/Kinetics:
- Astragalosides appear to exert their immunomodulatory actions via direct interaction with cellular membrane receptors. Shao et al. reported that astragalus root polysaccharide fractions activated mouse B cells and stimulated macrophage production via direct interaction between the polysaccharide fractions and TLR4 on cell surfaces.35 Hao et al. demonstrated in rat endothelial cells that astragalus root polysaccharide fractions stimulated the adhesion between neutrophils and endothelial cells by promoting the expression of superficial I-1 on the cell surface.87 This action would hypothetically facilitate the inflammatory response in wound healing and possibly underlie the detoxification and cellular protective actions of astragalus polysaccharides observed in several tissues. Astragalus polysaccharides also are known to increase RNA activity and protein synthesis in various tissues and increase the activity of enzymes that regulate specific cellular metabolic functions.2 Astragalus has been shown to modulate ornithine decarboxylase activity and increase insulin-induced protein tyrosine phosphorylase activity in the livers and skeletal muscles of animal models of type 2 diabetes.34
- There is insufficient available data on the pharmacokinetic parameters for individual chemical constituents of astragalus.
References
- Ko, J. K., Lam, F. Y., and Cheung, A. P. Amelioration of experimental colitis by Astragalus membranaceus through anti-oxidation and inhibition of adhesion molecule synthesis. World J Gastroenterol 10-7-2005;11(37):5787-5794. 16270386
- Zee-Cheng RK. Shi-quan-da-bu-tang (ten significant tonic decoction), SQT. A potent Chinese biological response modifier in cancer immunotherapy, potentiation and detoxification of anticancer drugs. Methods Find Exp Clin Pharmacol 1992;14(9):725-736. 1294861
- Zhao, K. S., Mancini, C., and Doria, G. Enhancement of the immune response in mice by Astragalus membranaceus extracts. Immunopharmacology 1990;20(3):225-233. 2149722
- Wang, H. Y. and Chen, Y. P. [Clinical observation on treatment of diabetic nephropathy with compound fructus arctii mixture]. Zhongguo Zhong Xi Yi Jie He Za Zhi 2004;24(7):589-592. 2149722
- Liu, Y. H., Yang, L., and Liu, J. [Clinical observation on treatment of early diabetic nephropathy by milkvetch injection combined with captopril]. Zhongguo Zhong Xi Yi Jie He Za Zhi 2005;25(11):993-995. 16355614
- Wu, Y., Ou-Yang, J. P., Wu, K., Wang, Y., Zhou, Y. F., and Wen, C. Y. Hypoglycemic effect of Astragalus polysaccharide and its effect on PTP1B. Acta Pharmacol Sin 2005;26(3):345-352. 15715932
- Zhang, D. Q., Zhang, J. J., Wang, J. X., Hu, S. M., Li, W., Ou, L. N., Chen, S. H., and Gao, X. M. [Effects of Qilan Tangzhining capsule on glucose and lipid metabolism in rats with diabetes mellitus and hyperlipemia]. Zhongguo Zhong Yao Za Zhi 2005;30(10):773-777. 16075719
- Shibata T, Sakai E, Nishi K, and Asetai M. Growth and glycoside content of Astragalus membranaceus (Bunge Leguminaceae) cultivated in different soil groups. Natural Medicine 1996;50:296-299.
- Qian, Z. W., Mao, S. J., Cai, X. C., Zhang, X. L., Gao, F. X., Lu, M. F., Shao, X. S., Li, Y. Y., Yang, X. K., Zhuo, Y. A., and . Viral etiology of chronic cervicitis and its therapeutic response to a recombinant interferon. Chin Med J (Engl) 1990;103(8):647-651. 2173655
- Yang Y, Jin P, Guo Q, and et al. Effect of Astragulas membranaceus on natural killer cell activity and induction of alpha- and gamma- interferon in patients with coxsackie B viral myocarditis. Chinese Medical Journal 1990;103(4):304-307.
- Li, C., Cao, L., and Zeng, Q. Astragalus prevents diabetic rats from developing cardiomyopathy by downregulating angiotensin II type2 receptors' expression. J Huazhong Univ Sci Technolog Med Sci 2004;24(4):379-384. 15587404
- Kim, C., Ha, H., Kim, J. S., Kim, Y. T., Kwon, S. C., and Park, S. W. Induction of growth hormone by the roots of Astragalus membranaceus in pituitary cell culture. Arch Pharm Res 2003;26(1):34-39. 12568355
- Ma, J., Peng, A., and Lin, S. Mechanisms of the therapeutic effect of astragalus membranaceus on sodium and water retention in experimental heart failure. Chin Med J (Engl) 1998;111(1):17-23. 10322646
- Guo, F. C., Kwakkel, R. P., Williams, B. A., Li, W. K., Li, H. S., Luo, J. Y., Li, X. P., Wei, Y. X., Yan, Z. T., and Verstegen, M. W. Effects of mushroom and herb polysaccharides, as alternatives for an antibiotic, on growth performance of broilers. Br Poult Sci 2004;45(5):684-694. 15623224
- Guo, F. C., Kwakkel, R. P., Williams, B. A., Parmentier, H. K., Li, W. K., Yang, Z. Q., and Verstegen, M. W. Effects of mushroom and herb polysaccharides on cellular and humoral immune responses of Eimeria tenella-infected chickens. Poult Sci 2004;83(7):1124-1132. 15285503
- Guo, F. C., Williams, B. A., Kwakkel, R. P., Li, H. S., Li, X. P., Luo, J. Y., Li, W. K., and Verstegen, M. W. Effects of mushroom and herb polysaccharides, as alternatives for an antibiotic, on the cecal microbial ecosystem in broiler chickens. Poult Sci 2004;83(2):175-182. 14979567
- Lu, D. C., Su, Z. J., and Rui, T. [Effect of jian yan ling on serum lipids, apoprotein and lipoprotein-a]. Zhongguo Zhong Xi Yi Jie He Za Zhi 1994;14(3):142-144, 131-2. 7950180
- Xu, C. J., Jian, X. C., and Cheng, H. Q. [Influence of astragalus membranaceus injection on the proliferation of rabbit bone marrow stromal cells and differentiation to osteoblast]. Zhong Nan Da Xue Xue Bao Yi Xue Ban 2004;29(4):489-491. 16134616
- Li, N. Q. [Clinical and experimental study on shen-qo injection with chemotherapy in the treatment of malignant tumor of digestive tract]. Zhongguo Zhong Xi Yi Jie He Za Zhi 1992;12(10):579, 588-592. 1302539
- Tianqing P, Yingzhen Y, Riesemann H, and et al. The inhibitory effect of Astragalus membranaceus on coxsackie B-3 virus RNA replication. Chinese Medical Sciences Journal 1995;10(3):146-150.
- Yan, H. J. [Clinical and experimental study of the effect of kang er xin-I on viral myocarditis]. Zhong Xi Yi Jie He Za Zhi 1991;11(8):468-70, 452. 1659497
- Yunde H, Guoliang M, Shuhua W, and et al. Effects of radix astragali seu-hedysari on teh interferon system. Chin Med J 1981;94:35-40.
- Kim SH, Lee SE, Oh H, and et al. The radioprotective effects of bu-zhong-yi-qi-tang: a prescription of traditional Chinesemedicine astragalus. J Chin Med 2002;30(1):127-137.
- Jia, W., Gao, W. Y., and Xiao, P. G. Antidiabetic drugs of plant origin used in China: compositions, pharmacology, and hypoglycemic mechanisms. Zhongguo Zhong Yao Za Zhi 2003;28(2):108-113. 15015278
- Xiao, H. B., Krucker, M., Albert, K., and Liang, X. M. Determination and identification of isoflavonoids in Radix astragali by matrix solid-phase dispersion extraction and high-performance liquid chromatography with photodiode array and mass spectrometric detection. J Chromatogr A 4-2-2004;1032(1-2):117-124. 15065786
- Wang, Q. J., Lu, W. W., Lu, H., Liu, F., Yang, S. J., Hua, Y. Q., and Ji, S. X. [Protective effect of qi dong yi xin on acute myocardial infarction in dogs]. Zhongguo Zhong Yao Za Zhi 2003;28(5):449-452. 15139132
- Kitagawa I, Wang H, Saito M, and et al. Chemical constituents of astragali radix, the root of Astragalus membranaceus bunge. Astragalosides, I, II, IV, acetylastragoloside I and isoastragalosides. Chem Pharm Bull 1983;31:698.
- He Z and Findlay JA. Constituents of Astragalus membranaceus. Journal of Natural Products 1991;54(3):810-815.
- Ma Y, Tian Z, Kuang H, and et al. Studies of the constituents of Astragalus membranaceus BUNGE. III. Structures of triterpenoidal glycosides, huangqiyenins A and B, from the leaves. Chem Pharm Bull 1997;45(2):359-361.
- Li, H. B., Ge, Y. K., Zhang, L., and Zheng, X. X. Astragaloside IV improved barrier dysfunction induced by acute high glucose in human umbilical vein endothelial cells. Life Sci 8-15-2006;79(12):1186-1193. 16650877
- Zhang, B. Q., Hu, S. J., Qiu, L. H., Zhu, J. H., Xie, X. J., Sun, J., Zhu, Z. H., Xia, Q., and Bian, K. Effects of Astragalus membranaceus and its main components on the acute phase endothelial dysfunction induced by homocysteine. Vascul Pharmacol 2007;46(4):278-285. 17196887
- Park JH, Kim BK, Park MK, and et al. Anti-diabetic activity of herbal drugs. Kor J Pharmacogn 1997;28(2):72-74.
- Shen, P., Liu, M. H., Ng, T. Y., Chan, Y. H., and Yong, E. L. Differential effects of isoflavones, from Astragalus membranaceus and Pueraria thomsonii, on the activation of PPARalpha, PPARgamma, and adipocyte differentiation in vitro. J Nutr 2006;136(4):899-905. 16549448
- Yong WU, Yand OU, Ping J, Ke WU, and et al. Hypoglycemic effect of astragalus polysaccharide and its effect on PTP1B. Acta Pharmacol Sin 2005;26(2):345-352.
- Shao, B. M., Xu, W., Dai, H., Tu, P., Li, Z., and Gao, X. M. A study on the immune receptors for polysaccharides from the roots of Astragalus membranaceus, a Chinese medicinal herb. Biochem Biophys Res Commun 8-6-2004;320(4):1103-1111. 15249203
- Zhang, H. M., Chen, S. W., Xie, C. G., Xie, Y. Q., Deng, X. F., and Wang, F. [Effects and mechanism of ShenQi compound recipe on inflammation maker in GK rats]. Zhong Yao Cai 2006;29(3):249-253. 16850723
- Zhang, W. J., Hufnagl, P., Binder, B. R., and Wojta, J. Antiinflammatory activity of astragaloside IV is mediated by inhibition of NF-kappaB activation and adhesion molecule expression. Thromb Haemost 2003;90(5):904-914. 14597987
- Hou, Y. D., Ma, G. L., Wu, S. H., Li, Y. Y., and Li, H. T. Effect of Radix Astragali seu Hedysari on the interferon system. Chin Med J (Engl) 1981;94(1):35-40. 6164521
- Kajimura, K., Takagi, Y., Ueba, N., Yamasaki, K., Sakagami, Y., Yokoyama, H., and Yoneda, K. Protective effect of astragali radix by oral administration against Japanese encephalitis virus infection in mice. Biol Pharm Bull 1996;19(9):1166-1169. 8889035
- Guo, Q., Peng, T. Q., and Yang, Y. Z. [Effect of Astragalus membranaceus on Ca2+ influx and coxsackie virus B3 RNA replication in cultured neonatal rat heart cells]. Zhongguo Zhong Xi Yi ie He Za Zhi 1995;15(8):483-485. 8580697
- Peng, T. Q., Yang, Y. Z., and Kandolf, R. [Effect and mechanism of Astragalus membranaceus on coxsackie B3 virus RNA in mice]. Zhongguo Zhong Xi Yi Jie He Za Zhi 1994;14(11):664-666. 7703635
- Yuan, W. L., Chen, H. Z., Yang, Y. Z., Yang, X. Y., Lin, Y. S., Jin, P. Y., and Zhou, T. S. Effect of astragalus membranaceus on electric activities of cultured rat beating heart cells infected with Coxsackie B-2 virus. Chin Med J (Engl) 1990;103(3):177-182. <2164463/a> 1371029
- Yao, X. J., Wainberg, M. A., and Parniak, M. A. Mechanism of inhibition of HIV-1 infection in vitro by purified extract of Prunella vulgaris. Virology 1992;187(1):56-62.
- Wei, H., Sun, R., Xiao, W., Feng, J., Zhen, C., Xu, X., and Tian, Z. Traditional Chinese medicine Astragalus reverses predominance of Th2 cytokines and their up-stream transcript factors in lung cancer patients. Oncol Rep 2003;10(5):1507-1512. 12883732
- Chu, D. T., Lepe-Zuniga, J., Wong, W. L., LaPushin, R., and Mavligit, G. M. Fractionated extract of Astragalus membranaceus, a Chinese medicinal herb, potentiates LAK cell cytotoxicity generated by a low dose of recombinant interleukin-2. J Clin Lab Immunol 1988;26(4):183-187. 3264344
- Chu, D., Sun, Y., Lin, J., Wong, W., and Mavligit, G. [F3, a fractionated extract of Astragalus membranaceus, potentiates lymphokine-activated killer cell cytotoxicity generated by low-dose recombinant interleukin-2]. Zhong Xi Yi Jie He Za Zhi 1990;10(1):34-6, 5. 2350835
- Chu, D. T., Lin, J. R., and Wong, W. [The in vitro potentiation of LAK cell cytotoxicity in cancer and aids patients induced by F3--a fractionated extract of Astragalus membranaceus]. Zhonghua Zhong Liu Za Zhi 1994;16(3):167-171. 7956691
- Jin, R., Wan, L. L., Mitsuishi, T., Kodama, K., and Kurashige, S. [Immunomodulative effects of Chinese herbs in mice treated with anti- tumor agent cyclophosphamide]. Yakugaku Zasshi 1994;114(7):533-538. 7932098
- Sun Y, Hersh EM, Talpax M, and et al. Immune restoration and/or augmentation of local graft versus host reaction by traditional Chinese medicinal herbs. Cancer 7-1-1983;52(1):70-73. 6336578
- Sun Y, Hersh EM, Lee S, and et al. Preliminary observations on the effects of the Chinese medicinal herbs Astragalus membranaceus and Ligustrum lucidum on lymphocyte blastogenic responses. Journal of Biological Response Modifiers 1983;2(3):227-237. 6644339
- Lau, B. H., Ruckle, H. C., Botolazzo, T., and Lui, P. D. Chinese medicinal herbs inhibit growth of murine renal cell carcinoma. Cancer Biother 1994;9(2):153-161. 7812364
- Wang, Q. [Inotropic action of Astragalus membranaceus Bge. saponins and its possible mechanism]. Zhongguo Zhong Yao Za Zhi 1992;17(9):557-559 (inside backcover). 1292500
- Wong, B. Y., Lau, B. H., Tadi, P. P., and Teel, R. W. Chinese medicinal herbs modulate mutagenesis, DNA binding and metabolism of aflatoxin B1. Mutat Res 6-1-1992;279(3):209-216. 1377337
- Shirataki Y, Takao M, Yoshida S, and et al. Antioxidative components isolated from the roots of Astragalus membranaceus Bunge (Astragali Radix) Phytotherapy Research 1997;11:603-605.
- Xu, X. Y., Li, L. H., Wu, L. S., Zhao, C. L., and Lin, H. Y. [Adjustment effect of Radix Astragalus and Radix Angelicae sinensis on TNF-alpha and bFGF on renal injury induced by ischemia reperfusion in rabbit]. Zhongguo Zhong Yao Za Zhi 2002;27(10):771-773. 12776561
- Sheng, B. W., Chen, X. F., Zhao, J., He, D. L., and Nan, X. Y. Astragalus membranaceus reduces free radical-mediated injury to renal tubules in rabbits receiving high-energy shock waves. Chin Med J (Engl) 1-5-2005;118(1):43-49. 15642225
- Guan, Y., Zhou, Z., Zhang, C., Ruan, Y., and Wu, Z. Effect of A-L tonic capsule on DNA content in rat experimental hepatocarcinogenesis. J Huazhong Univ Sci Technolog Med Sci 2002;22(1):12-3, 16. 12658771
- Han, L. and Chen, K. J. [The protective effects of yi-xin-kang capsule on the structure and function of rat myocardial mitochondria]. Zhongguo Zhong Yao Za Zhi 2001;26(11):773-777. 12776352
- Hong CY, Lo YC, Tan FC, and et al. Astragalus membranaceus and Polygonum multiflorum protect rat heart mitochondria against lipid peroxidation. American Journal of Chinese Medicine 1994;22(1):63-70. 8030620
- Toda, S., Yase, Y., and Shirataki, Y. Inhibitory effects of astragali radix, crude drug in Oriental medicines on lipid peroxidation and protein oxidative modification of mouse brain homogenate by copper. Phytother Res 2000;14(4):294-296. 10861978
- Wang, D., Shen, W., Tian, Y., Dong, Z., Liu, G., Sun, Z., Yang, S., and Zhou, S. [Protective effect of total flavonoids of radix Astragali on mammalian cell damage caused by hydroxyl radical]. Zhongguo Zhong Yao Za Zhi 1995;20(4):240-2. 7646797
- Wang, D. Q., Critchley, J. A., Ding, B. G., Ma, Y. Q., Zhao, H. L., Thomas, G. N., Tian, Y. P., Tomlinson, B., Critchley, L. A., and Edward, J. A. [Protection against paracetamol-induced hepatic damage using total flavonoids of Astragalus]. Zhongguo Zhong Yao Za Zhi 2001;26(9):617-620. 12776431
- Wang, D. Q., Ding, B. G., Ma, Y. Q., Zhao, H. L., Neil, T. G., Brian, T., Tian, Y. P., Wang, C. B., and Critchley, J. A. [Studies on protective effect of total flavonoids of Astragalus on liver damage paracetamol-inducedby]. Zhongguo Zhong Yao Za Zhi 2001;26(7):483-486. 12776364
- Wu, L., Liu, H., Xue, P., Lu, Z. G., and Du, K. F. [Influence of a triplex superimposed treatment on HBV replication and mutation during treating chronic hepatitis B]. Zhonghua Shi YanHe Lin Chuang Bing Du Xue Za Zhi 2001;15(3):236-238. 11986694
- Xuan, W., Dong, M., and Dong, M. Effects of compound injection of Pyrola rotundifolia L and Astragalus membranaceus Bge on experimental guinea pigs' gentamicin ototoxicity. Ann Otol Rhinol Laryngol 1995;104(5):374-380. 7747908
- Zhou, J. Y., Fan, Y., Kong, J. L., Wu, D. Z., and Hu, Z. B. [Effects of components isolated from Astragalus membranaceus Bunge on cardiac function injured by myocardial ischemia reperfusion in rats]. Zhongguo Zhong Yao Za Zhi 2000;25(5):300-302. 12512456
- Hei, Z. Q., Zhang, J. J., Lin, S. Q., Liu, K. X., and Chen, B. X. [Effects of Astragalus membranaceus injection on nitric oxide and endothelin concentration of intestinal mucosa after hemorrhage shock-reperfusion in rats]. Zhongguo Zhong Yao Za Zhi 2004;29(5):444-447. 15706900
- He, X., Li, C., and Yu, S. Protective effects of radix astragali against anoxic damages to in vitro cultured neurons. J Tongji Med Univ 2000;20(2):126-127. 12845724
- Luo, Y., Qin, Z., Hong, Z., Zhang, X., Ding, D., Fu, J. H., Zhang, W. D., and Chen, J. Astragaloside IV protects against ischemic brain injury in a murine model of transient focal ischemia. Neurosci Lett 6-17-2004;363(3):218-223. 15182947
- Zhang WJ, Wojta J, and Binder BR. Regulation of the fibrinolytic potential of cultured human umbilical vein endothelial cells: Astragaloside IV downregulates plasminogen activator inhibitor-1 and upregulates tissue-type plasminogen activator expression. J Vasc Res 1997;34:273-280.
- Hikino H, Funayama S, and Endo K. Hypotensive principle of Astragalus and Hedysarum roots. Planta Medica 1976;30:297-302.
- Ning, L., Chen, C. X., Jin, R. M., Wu, Y. P., Zhang, H. G., Sun, C. L., Song, C. Q., and Hu, Z. B. [Effect of components of dang-gui-bu-xue decoction on hematopenia]. Zhongguo Zhong Yao Za Zhi 2002;27(1):50-53. 12774358
- Chen, M. L., Liu, W. J., Wang, C. Y., Zhu, X. M., Yin, J. F., Wang, D. X., and Liu, P. [Effects of yishenjiangyafang on blood pressure and protecting renal function in RPH rats]. Zhongguo Zhong Yao Za Zhi 2003;28(8):746-751. 15015358
- Lei, Y., Gao, Q., and Li, Y. S. [Study on effects of Astragalus, Angelica and their combination on vascular endothelial cell proliferation in vitro]. Zhongguo Zhong Xi Yi Jie He Za Zhi 2003;23(10):753-756. 14626189
- Jin, C. and Dai, R. H. [Effect of Astragalus membranaceus on erythrocyte sodium content and sodium transport in the coronary heart disease]. Zhong Xi Yi Jie He Za Zhi 1991;11(11):651-3, 643. 1667511
- Kim, C., Ha, H., Lee, J. H., Kim, J. S., Song, K., and Park, S. W. Herbal extract prevents bone loss in ovariectomized rats. Arch Pharm Res 2003;26(11):917-924. 14661857
- Hong, C. Y., Ku, J., and Wu, P. Astragalus membranaceus stimulates human sperm motility in vitro. Am J Chin Med 1992;20(3-4):289-294. 1471613
- Liu, J., Liang, P., Yin, C., Wang, T., Li, H., Li, Y., and Ye, Z. Effects of several Chinese herbal aqueous extracts on human sperm motility in vitro. Andrologia 2004;36(2):78-83. 15084153
- Zhuang MX. The effects of polysaccharides of Astragalus membranaceus Codonopsis pilosula and Panax ginseng on some immune functions in guinea-pigs. Zhongguo yaoxue zazhi 1992;27:653-655.
- He, J., Li, Y., Wei, S., Guo, M., and Fu, W. [Effects of mixture of Astragalus membranaceus, Fructus Ligustri lucidi and Eclipta prostrata on immune function in mice]. Hua Xi Yi e Da Xue Xue Bao 1992;23(4):408-411. 1304546
- Jiao, Y., Wen, J., and Yu, X. [Influence of flavonoid of Astragalus membranaceus's stem and leaves on the function of cell mediated immunity in mice]. Zhongguo Zhong Xi Yi Jie He Za Zhi 1999;19(6):356-358. 11783202
- Liang, H., Zhang, Y., and Geng, B. [The effect of astragalus polysaccharides (APS) on cell mediated immunity (CMI) in burned mice]. Zhonghua Zheng Xing Shao Shang Wai Ke Za Zhi 1994;10(2):138-141. 7922816
- Yoshida, Y., Wang, M. Q., Liu, J. N., Shan, B. E., and Yamashita, U. Immunomodulating activity of Chinese medicinal herbs and Oldenlandia diffusa in particular. Int J Immunopharmacol 1997;19(7):359-370. 9568540
- Zhao, K. W. and Kong, H. Y. [Effect of Astragalan on secretion of tumor necrosis factors in human peripheral blood mononuclear cells]. Zhongguo Zhong Xi Yi Jie He Za Zhi 1993;13(5):263-5, 259. 8219673
- Lau BH, Ong P, and Tosk J. Macrophage chemiluminescence modulated by Chinese medicinal herbs Astragalus membranaceus and Ligustrum lucidum. Phytotherapy Research 1989;3(4):148-153.
- Yang, B. H., Zhu, L. Q., Zhang, J. Z., Niu, F. L., and Cui, W. [Effects of Astragalus membranaceus and Panax notoginseng on the transformation of bone marrow stem cells and proliferation of EPC in vitro]. Zhongguo Zhong Yao Za Zhi 2005;30(22):1761-1763. 16468374
- Hao, Y., Qiu, Q. Y., and Wu, J. [Effect of Astragalus polysaccharides in promoting neutrophil-vascular endothelial cell adhesion and expression of related adhesive molecules]. Zhongguo Zhong Xi Yi Jie He Za Zhi 2004;24(5):427-430. 15199628
Licensed by Natural Standard Copyright © 2010 by Natural Standard Corporation. All Rights Reserved.
back to Plant Profiler back to top
|