Sigma-Aldrich
Life Science > Nutrition Research > Learning Center > Plant Profiler > Black cohosh (Cimicifuga racemosa)
Plant Profiler
Black cohosh (Cimicifuga racemosa)
Synonyms / Common Names / Related Terms
Actaea macrotys, Actaea pachypoda, Actaea podocarpa, Actaea racemosa L., Actaea rubra actaealactone, actee a grappes (French), amerikanisches Wanzenkraut (German), Appalachian bugbane, BCE, baneberry, black bugbane, black cohosh root extract Cr 99, black cohosh roots, black snakeroot, botrophis serpentaria, bugbane, bugwort, CR, CR BNO 1055, CR extract, caffeic acid, cohosh bugbane, Cimicifuga, Cimicifuga racemosa, Cimicifugae racemosae rhizoma, Cimicifugawurzelstock, cimicifugic acid A, cimicifugic acid B, cimicifugic acid D, cimicifugic acid E, cimicifugic acid F, cimicifugic acid G, cohosh bugbane, ferulic acid, fukinolic acid, herbe au punaise (French), ICR, isoferulic acid, isopropanolic black cohosh extract, isopropanolic extract, macrotys, Macrotys actaeoides, methyl caffeate, mountain bugbane, p-coumaric acid, phytoestrogen, protocatechualdehyde, protocatechuic acid, Ranunculaceae (family), rattle root, rattle snakeroot, rattlesnake root, rattle top, rattle weed, rattleweed, Remifemin®, rhizoma actaeae, rich weed, richweed, schwarze Schlangenwurzel (German), snakeroot, solvlys, squaw root, squawroot, Thalictrodes racemosa, Traubensilberkerze (German), triterpene glycosides, Wanzenkraut, Ze 450.
Bioactive products found in Cimicifuga racemosa
Mechanism of Action
Pharmacology:
  • Constituents: Constituents of black cohosh with proposed or demonstrated pharmacological activity include triterpine glycosides (23-epi-26-deoxyactein, actein, 27-deoxyactein, cimicifugoside A, cimicifugoside M, cimiracemoside A-H)21,45,46,47,48,12,49,50,51, cyclolanostanol xylosides 52, isoflavonone formononetin 53, hydroxytyrosol 54, actaeaeposcide 55, phenylpropanoids (cimiracemate A, cimiracemate B)8,56, organic acids (caffeic acid, cimicifugic acid A, cimicifugic acid B, cimicifugic acid E, cimicifugic acid F, cinnamic acid ester dehydrocimicifugic A, dehydrocimicifugic acid B, dihydroxyphenyl lactic acid, ferulic acid, fukinolic acid, isoferulic acid, methyl caffeate acid, salicylic acid)8,57,54,58, phenols57, quinoid metabolites of caffeic acid, cimiracemate B, fukinolic acid, fukiic acid, and.hydroxytyrosol54. Actaea racemosa specifically contains lignans (e.g., actaealactone), phenylpropanoid ester derivatives (e.g., cimicifugic acid), polyphenols (0.36-2.92% (w/w) in dried root and rhizome), protocatechuic acid, protocatechualdehyde, p-coumaric acid, caffeic acid, methyl caffeate, ferulic acid, ferulate-1-methyl ester, isoferulic acid, 1-isoferuloyl-beta-d-glucopyranoside, fukinolic acid, and cimicifugic acids A, B, and D-F.9,59. Methods of extracting active ingredients from black cohosh have been described.60
  • Anti-climacteric effects: According to in vitro study, black cohosh was found to be consistent with a human mu opiate receptor (hMOR) agonist, with an EC50 of 68.8 ± 7.7mcg/mL, which may explain its purported beneficial role in alleviating menopausal symptoms.61
  • Anti-coagulation effects: Native black cohosh contains small amounts of salicylic acid, but it is not clear how much (if any) is present in commercially available or standardized extracts. A popular combination product, Reumalex®, contains black cohosh, white willow bark, sarsaparilla, poplar bark, and guaiacum resin. Notably, several constituents contain salicylates, and it has been estimated that each Reumalex® tablet may include up to 10-20mg of salicylates.
  • Anti-inflammatory activity: Ovariectomized Wistar rats administered a phytoestrogen compound (genistein, daidzein, glycitein, black cohosh, Angelica, licorice, and Vitex agnus-castus) orally showed a significantly lower level of proinflammatory cytokines and a higher level of TGF-beta.38 The constituent isoferulic acid has also been reported to have anti-inflammatory effects and may decrease muscular spasm.11 Furthermore, salicylic acid is found in small quantities in black cohosh, and it is presumed that the salicylic acid contributes to the anti-inflammatory and analgesic properties of black cohosh.
  • Antineoplastic effects: Based on a systematic review, there is laboratory evidence of antiproliferative properties of black cohosh but a lack of confirmation from clinical studies for a protective role in cancer prevention.19 In vitro studies have reported black cohosh to possess inhibitory effects on estrogen responsive cancer cell lines/breast cancer cells.37,45,62,63,22,21 Furthermore, in a cell line study, relatively low concentrations of actein or the methanol/water fraction of black cohosh may cause synergistic inhibition of human breast cancer cell proliferation when combined with different classes of chemotherapy agents.12 Acetein may activate genes that respond to DNA damage and unfolded protein responses, and enhance apoptosis and repressed cell cycle genes.64,65 Actaealactone and cimicifugic acid may also have a small stimulating effect on the growth of breast cancer cell proliferation.9 In an in vitro study conducted on human prostate cancer cells with black cohosh extract, the extract killed hormone-responsive or hormone-unresponsive prostate cancer cells by induction of apoptosis and activation of caspases.13 Another in vivo study in mice demonstrated inhibited PC3 prostate cancer tumor growth with black cohosh and other herbal extracts.14 The mechanism behind tumor inhibition appeared to be anti-angiogenic by decreasing intratumoral microvessel density.
  • Antioxidant effects: Extracts of black cohosh have protected against induced DNA damage through scavenging of reactive oxygen species in vitro.8 A sample of black cohosh collected in 1919 by the physician and plant explorer Henry Hurd Rusby, was recently identified in the collections of The New York Botanical Garden and analyzed. A comparison of the triterpene glycosidic and phenolic constituents of the 85 year-old plant sample with those of a modern collection of Actaea racemosa showed the similarity of the two samples, and both extracts showed similar antioxidant activity. This confirms the stability of the older sample, despite curation.44 In laboratory study, the constituents, actaealactone, cimicifugic acid, and fukinolic acid may display antioxidant activity in the 1,1-diphenyl-2-picrylhydrazyl (DPPH) free-radical assay with IC50 values of 26 and 37mcM, respectively.9,59
  • Bone metabolism effects: An isopropanolic extract of black cohosh has been shown to significantly diminish the urinary content of pyridinoline and deoxypyridinoline, specific markers for bone loss, and the morphometric correlates of bone loss associated with ovariectomy in rats.15 Ovariectomized rats treated with Cimicifuga racemosa extract had slightly stimulated gene expression of IGF-I, collagen-lαl, osteoprotegerin and osteocalcin (all osteoblast products), and of tartrate-resistant acid phophatas (TRAP, an osteoclast product) in the metaphysic of the femur.6 In an in vitro study, an isopropanolic extract (iCR) from the rhizomes of black cohosh stimulated osteoblastic osteoprotegerin protein secretion by 3- to 5-fold as early as 12h without affecting receptor activator for nuclear factor κB ligand (RANKL) expression.42
  • CNS effects: Recent studies suggest that the mechanism of action of black cohosh may be centrally mediated, with possible action at the level of serotonin or dopamine receptors.2,3 A study in ovariectomized rats demonstrated strong binding to serotonin receptors 5-HT(1A), 5-HT(1D), and 5-HT(7) subtypes.2
  • Cytochrome P450 (CYP) effects: In a clinical trial of 12 healthy volunteers (six women), black cohosh daily for 28 days had a statistically significant inhibition of CYP2D6 (difference, -0.046; 95% CI, -0.085 to -0.007), but the magnitude of the effect (approximately 7%) did not appear to be clinically relevant.40 In another trial by the same authors, healthy volunteers taking black cohosh 80mg daily for 14 days did not have a clinically relevant effect on their CYP3A activity.39
  • Endocrine effects: Cimicifugoside contained in black cohosh is believed to affect hypothalamus-pituitary function.
  • Estrogenic effects: It is not clear what constituent(s) of black cohosh, if any, possesses estrogenic properties. In animals and in vitro, initial reports of estrogen receptor binding activity.27,30 stand in contrast with more recent data suggesting no significant estrogen receptor binding activity or estrogenic activities.66,1,4,29,30,20,5,67,68,46,28,10 Two in vitro studies found no effects of black cohosh alone on estrogen receptors, but reported that black cohosh antagonized proliferative effects on cells induced by estradiol.31,31 A similar in vitro study on estrogen sensitive breast cancer cells (MCF-7) reported isopropanolic black cohosh extract did not stimulate MCF-7 growth and exerted inhibitory effects on cellular proliferation, indicating strong estrogen-antagonist effects.69 The proliferation-inhibiting effect of tamoxifen has been enhanced by black cohosh extract.31 Several studies have aimed to assess estrogen activity by measuring luteinizing hormone (LH), follicle stimulating hormone (FSH), or prolactin levels.32,33 One study reported lower FSH levels (but not LH) in patients treated with black cohosh vs. placebo (N=110), although baseline hormone levels were not known in either group.32 Results from other trials have found no effects on these hormone levels after up to six months of black cohosh therapy.34,17,23 Administered to female infantile mice, premature onset of estrus could not be precipitated by black cohosh.35 Estrogenic effects on vaginal epithelium were noted in one three-month trial of black cohosh36, while a more recent six-month trial reported no effects on vaginal cytology18. Cimicifuga racemosa ethanolic extract, Ze 450, may inhibit cell proliferation and show antiestrogenic activity.70 The Cimicifuga racemosa extract bound to the progesterone receptor B1 but did not show progestin-like activity in the T-47D cell line. In an in vitro study using healthy breast tissue of pre- and postmenopausal women, incubation in vitro with black cohosh extract decreased local estrogen formation.71
  • Gastrointestinal effects: In an in vitro study, black cohosh extracts moderately (but significantly) inhibited estrone-3-sulfate (a typical organic anion-transporting polypeptide B (OATP-B) substrate) uptake by 47.2% (p<0.05).41 As OAT-B is involved in the intestinal absorption of various drugs, black cohosh may decrease the absorption of orally administered substrates of OATP-B.
  • Neuropharmacologic effects: Black cohosh has been shown to exhibit an action on the central endogenous opioid system in postmenopausal women as evidenced by suppression of mean luteinizing hormone pulse frequency following opioid receptor blockade.72
  • Serum glucose level altering effects: In a randomized trial in 351 peri- or post-menopausal women, black cohosh had no demonstrable effects on lipids, glucose, insulin, or fibrinogen.24
  • Serum insulin level altering effects: In a randomized trial in 351 peri- or post-menopausal women, black cohosh had no demonstrable effects on lipids, glucose, insulin, or fibrinogen.24
  • Serum lipid level altering effects: In a double-blind randomized, placebo controlled, study in 89 peri- or postmenopausal women experiencing climacteric symptoms, a combination of black cohosh (Cimicifuga racemosa) and St. John's wort (Hypericum perforatum) significantly increased HDL levels (from 58.32 ± 11.64 to 59.74 ± 10.54; p=0.04) compared to the control (from 60.20 ± 16.37 to 56.63 ± 12.67).25 However, in a randomized trial in 351 peri- or post-menopausal women, black cohosh had no demonstrable effects on lipids, glucose, insulin, or fibrinogen.24
  • Vascular effects: In a 1962 study, acteina, a constituent of black cohosh, was found to cause peripheral vasodilation, and has been noted to elicit hypotension in animals.26 Additional supporting data in humans is lacking.
  • Vasoactive effects: Laboratory study of cimicifugic acids C and D, and fukinolic acid in the rhizome of black cohosh show vasoactive effects.43
  • Other: In vivo oral administration of black cohosh extract inhibited the anti-IgE-induced passive cutaneous anaphylaxis reaction.7 Black cohosh extract also showed inhibitory potential on histamine release. Cimicifugoside from Cimicifuga simplex has been found to inhibit cellular thymidine-3H uptake, and to act as a selective inhibitor of nucleoside transport into mammalian cells.73,74,16,73
Pharmacokinetics/Pharmacodynamics:
  • According to in vitro study, black cohosh was found to be consistent with a human mu opiate receptor (hMOR) agonist, with an EC50 of 68.8 ± 7.7mcg/mL, which may explain its purported beneficial role in alleviating menopausal symptoms.61
  • In a clinical trial, oral black cohosh administration for 14 days did not significantly affect digoxin's area under the serum concentration time curves from 0-3 hours (AUC(0-3)), AUC(0-24), Cmax, apparent oral clearance of digoxin (CL/F), and elimination half-life.75 Digoxin is a known P-glycoprotein (P-gp) substrate, which suggests that black cohosh is not a potent modulator of P-gp in vivo.
References
  1. Borrelli, F., Izzo, A. A., and Ernst, E. Pharmacological effects of Cimicifuga racemosa. Life Sci 7-25-2003;73(10):1215-1229. 12850238
  2. Burdette, J. E., Liu, J., Chen, S. N., Fabricant, D. S., Piersen, C. E., Barker, E. L., Pezzuto, J. M., Mesecar, A., van Breemen, R. B., Farnsworth, N. R., and Bolton, J. L. Black cohosh acts as a mixed competitive ligand and partial agonist of the serotonin receptor. J Agric Food Chem 9-10-2003;51(19):5661-5670. 12952416
  3. Jarry, H., Metten, M., Spengler, B., Christoffel, V., and Wuttke, W. In vitro effects of the Cimicifuga racemosa extract BNO 1055. Maturitas 3-14-2003;44 Suppl 1:S31-S38. 12609557
  4. Lupu, R., Mehmi, I., Atlas, E., Tsai, M. S., Pisha, E., Oketch-Rabah, H. A., Nuntanakorn, P., Kennelly, E. J., and Kronenberg, F. Black cohosh, a menopausal remedy, does not have estrogenic activity and does not promote breast cancer cell growth. Int J Oncol 2003;23(5):1407-1412. 14532983
  5. Mahady, G. B. Is black cohosh estrogenic? Nutr.Rev 2003;61(5 Pt 1):183-186. 12822708
  6. Seidlova-Wuttke, D., Hesse, O., Jarry, H., Christoffel, V., Spengler, B., Becker, T., and Wuttke, W. Evidence for selective estrogen receptor modulator activity in a black cohosh (Cimicifuga racemosa) extract: comparison with estradiol-17beta. Eur J Endocrinol 2003;149(4):351-362. 14514351
  7. Kim, C. D., Lee, W. K., Lee, M. H., Cho, H. S., Lee, Y. K., and Roh, S. S. Inhibition of mast cell-dependent allergy reaction by extract of black cohosh (Cimicifuga racemosa). Immunopharmacol Immunotoxicol 2004;26(2):299-308. 15209365
  8. Burdette, J. E., Chen, S. N., Lu, Z. Z., Xu, H., White, B. E., Fabricant, D. S., Liu, J., Fong, H. H., Farnsworth, N. R., Constantinou, A. I., van Breemen, R. B., Pezzuto, J. M., and Bolton, J. L. Black cohosh (Cimicifuga racemosa L.) protects against menadione-induced DNA damage through scavenging of reactive oxygen species: bioassay-directed isolation and characterization of active principles. J Agric Food Chem 11-20-2002;50(24):7022-7028. 12428954
  9. Nuntanakorn, P., Jiang, B., Einbond, L. S., Yang, H., Kronenberg, F., Weinstein, I. B., and Kennelly, E. J. Polyphenolic constituents of Actaea racemosa. J Nat Prod 2006;69(3):314-318. 16562825
  10. Zhang, L., Khan, I. A., Willett, K. L., and Foran, C. M. In vivo effects of black cohosh and genistein on estrogenic activity and lipid peroxidation in Japanese Medaka (Oryzias latipes). J Herb Pharmacother 2003;3(3):33-50. 15277055
  11. Shibata, M., Ikoma, M., Onoda, M., Sato, F., and Sakurai, N. [Pharmacological studies on the Chinese crude drug "Shoma". III. Central depressant and antispasmodic actions of Cimicifuga rhizoma, Cimicifuga simplex Wormsk (author's transl)]. Yakugaku Zasshi 1980;100(11):1143-1150. 7205558
  12. Einbond, L. S., Shimizu, M., Nuntanakorn, P., Seter, C., Cheng, R., Jiang, B., Kronenberg, F., Kennelly, E. J., and Weinstein, I. B. Actein and a fraction of black cohosh potentiate antiproliferative effects of chemotherapy agents on human breast cancer cells. Planta Med 2006;72(13):1200-1206. 16988912
  13. Hostanska, K., Nisslein, T., Freudenstein, J., Reichling, J., and Saller, R. Apoptosis of human prostate androgen-dependent and -independent carcinoma cells induced by an isopropanolic extract of black cohosh involves degradation of cytokeratin (CK) 18. Anticancer Res 2005;25(1A):139-147. 15816531
  14. Ng, S. S. and Figg, W. D. Antitumor activity of herbal supplements in human prostate cancer xenografts implanted in immunodeficient mice. Anticancer Res 2003;23(5A):3585-3590. 14666653
  15. Nisslein, T. and Freudenstein, J. Effects of an isopropanolic extract of Cimicifuga racemosa on urinary crosslinks and other parameters of bone quality in an ovariectomized rat model of osteoporosis. J Bone Miner Metab 2003;21(6):370-376. 14586793
  16. Takahira, M., Kusano, A., Shibano, M., Kusano, G., Koizumi, K., Suzuki, R., Kim, H. S., Wataya, Y., and Takahara, M. Antimalarial activity and nucleoside transport inhibitory activity of the triterpenic constituents of Cimicifuga spp. Biol Pharm Bull 1998;21(8):823-828. 9743250
  17. Lehmann-Willenbrock, E. and Riedel, H. H. [Clinical and endocrinologic studies of the treatment of ovarian insufficiency manifestations following hysterectomy with intact adnexa]. Zentralbl Gynakol 1988;110(10):611-618. 2841818
  18. Liske E and Wüstenberg P. Therapy of climacteric complaints with Cimicifuga racemosa: herbal medicine with clinically proven evidence [poster presentation]. Menopause 1998;5(4):250.
  19. Walji, R., Boon, H., Guns, E., Oneschuk, D., and Younus, J. Black cohosh (Cimicifuga racemosa [L.] Nutt.): safety and efficacy for cancer patients. Support Care Cancer 2007;15(8):913-921. 17602247
  20. Einer-Jensen, N., Zhao, J., Andersen, K. P., and Kristoffersen, K. Cimicifuga and Melbrosia lack oestrogenic effects in mice and rats. Maturitas 1996;25(2):149-153. 8905606
  21. Struck D, Tegtmeier M, and Harnischfeger G. Flavones in extracts of Cimicifuga racemosa. Planta Med 1997;63:289-290.
  22. Nesselhut T, Schellhase C, Dietrich R, and et al. [Investigations into the growth-inhibitive efficacy of phytopharmacopia with estrogen-like influences on mammary gland carcinoma cells] (translated from German). Arch Gynecol Obstet 1993;254:817-818.
  23. Jacobson, J. S., Troxel, A. B., Evans, J., Klaus, L., Vahdat, L., Kinne, D., Lo, K. M., Moore, A., Rosenman, P. J., Kaufman, E. L., Neugut, A. I., and Grann, V. R. Randomized trial of black cohosh for the treatment of hot flashes among women with a history of breast cancer. J Clin Oncol 5-15-2001;19(10):2739-2745. 11352967
  24. Spangler, L., Newton, K. M., Grothaus, L. C., Reed, S. D., Ehrlich, K., and LaCroix, A. Z. The effects of black cohosh therapies on lipids, fibrinogen, glucose and insulin. Maturitas 6-20-2007;57(2):195-204. 17275226
  25. Chung, D. J., Kim, H. Y., Park, K. H., Jeong, K. A., Lee, S. K., Lee, Y. I., Hur, S. E., Cho, M. S., Lee, B. S., Bai, S. W., Kim, C. M., Cho, S. H., Hwang, J. Y., and Park, J. H. Black cohosh and St. John's wort (GYNO-Plus) for climacteric symptoms. Yonsei Med J 4-30-2007;48(2):289-294. 17461529
  26. Genazzani E and Sorrentino L. Vascular action of acteina: active constituent of Actaea racemosa L. Nature 1962;194(4828):544-545.
  27. Jarry, H., Harnischfeger, G., and Duker, E. [The endocrine effects of constituents of Cimicifuga racemosa. 2. In vitro binding of constituents to estrogen receptors]. Planta Med 1985;51(4):316-319. 4070446
  28. Zava, D. T., Dollbaum, C. M., and Blen, M. Estrogen and progestin bioactivity of foods, herbs, and spices. Proc Soc Exp Biol Med 1998;217(3):369-378. 9492350
  29. Liu, J., Burdette, J. E., Xu, H., Gu, C., van Breemen, R. B., Bhat, K. P., Booth, N., Constantinou, A. I., Pezzuto, J. M., Fong, H. H., Farnsworth, N. R., and Bolton, J. L. Evaluation of estrogenic activity of plant extracts for the potential treatment of menopausal symptoms. J Agric Food Chem 2001;49(5):2472-2479. 11368622
  30. Liu, Z., Yang, Z., Zhu, M., and Huo, J. [Estrogenicity of black cohosh (Cimicifuga racemosa) and its effect on estrogen receptor level in human breast cancer MCF-7 cells]. Wei Sheng Yan Jiu 2001;30(2):77-80. 11321955
  31. Zierau, O., Bodinet, C., Kolba, S., Wulf, M., and Vollmer, G. Antiestrogenic activities of Cimicifuga racemosa extracts. J Steroid Biochem Mol Biol 2002;80(1):125-130. 11867271
  32. Duker, E. M., Kopanski, L., Jarry, H., and Wuttke, W. Effects of extracts from Cimicifuga racemosa on gonadotropin release in menopausal women and ovariectomized rats. Planta Med 1991;57(5):420-424. 1798794
  33. Jarry, H. and Harnischfeger, G. [Endocrine effects of constituents of Cimicifuga racemosa. 1. The effect on serum levels of pituitary hormones in ovariectomized rats]. Planta Med 1985;51(1):46-49. 3925475
  34. Liske E, Wüstenberg P, and Boblitz N. Human-pharmacological investigations during treatment of climacteric complaints with Cimicifuga racemosa (Remifemin): No estrogen-like effects. ESCOP 2001;1:1.
  35. Siess VM and Seybold G. [Studies on the effects of Pulsatilla pratensis, Cimicifuga racemosa and Aristolochia clematitis on the estrus in infantile and castrated white mice]. Arzneimittelforschung 1960;10:514-520.
  36. Stoll W. Phytotherapeutikum beeinflusst atrophisches Vaginal epithel. Doppelblindversuch Cimicifuga vs. Oestrogenpraeparat [Phytopharmaceutical influences on atrophic vaginal epithelium. Double-blind study on Cimicifuga vs. an estrogen preparation]. Therapeutickon 1987;1:23-32.
  37. Dixon-Shanies, D. and Shaikh, N. Growth inhibition of human breast cancer cells by herbs and phytoestrogens. Oncol Rep 1999;6(6):1383-1387. 10523716
  38. Marotta, F., Mao, G. S., Liu, T., Chui, D. H., Lorenzetti, A., Xiao, Y., and Marandola, P. Anti-inflammatory and neuroprotective effect of a phytoestrogen compound on rat microglia. Ann N Y Acad Sci 2006;1089:276-281. 17261775
  39. Gurley, B., Hubbard, M. A., Williams, D. K., Thaden, J., Tong, Y., Gentry, W. B., Breen, P., Carrier, D. J., and Cheboyina, S. Assessing the clinical significance of botanical supplementation on human cytochrome P450 3A activity: comparison of a milk thistle and black cohosh product to rifampin and clarithromycin. J Clin Pharmacol 2006;46(2):201-213. 16432272
  40. Gurley, B. J., Gardner, S. F., Hubbard, M. A., Williams, D. K., Gentry, W. B., Khan, I. A., and Shah, A. In vivo effects of goldenseal, kava kava, black cohosh, and valerian on human cytochrome P450 1A2, 2D6, 2E1, and 3A4/5 phenotypes. Clin Pharmacol Ther 2005;77(5):415-426. 15900287
  41. Fuchikami, H., Satoh, H., Tsujimoto, M., Ohdo, S., Ohtani, H., and Sawada, Y. Effects of herbal extracts on the function of human organic anion-transporting polypeptide OATP-B. Drug Metab Dispos 2006;34(4):577-582. 16415120
  42. Viereck, V., Grundker, C., Friess, S. C., Frosch, K. H., Raddatz, D., Schoppet, M., Nisslein, T., Emons, G., and Hofbauer, L. C. Isopropanolic extract of black cohosh stimulates osteoprotegerin production by human osteoblasts. J Bone Miner Res 2005;20(11):2036-2043. 16234977
  43. Noguchi, M., Nagai, M., Koeda, M., Nakayama, S., Sakurai, N., Takahira, M., and Kusano, G. Vasoactive effects of cimicifugic acids C and D, and fukinolic acid in cimicifuga rhizome. Biol Pharm Bull 1998;21(11):1163-1168. 9853406
  44. Jiang, B., Yang, H., Nuntanakorn, P., Balick, M. J., Kronenberg, F., and Kennelly, E. J. The value of plant collections in ethnopharmacology: a case study of an 85-year-old black cohosh (Actaea racemosa L.) sample. J Ethnopharmacol 1-15-2005;96(3):521-528. 15619573
  45. Einbond, L. S., Shimizu, M., Xiao, D., Nuntanakorn, P., Lim, J. T., Suzui, M., Seter, C., Pertel, T., Kennelly, E. J., Kronenberg, F., and Weinstein, I. B. Growth inhibitory activity of extracts and purified components of black cohosh on human breast cancer cells. Breast Cancer Res Treat 2004;83(3):221-231. 14758092
  46. Onorato, J. and Henion, J. D. Evaluation of triterpene glycoside estrogenic activity using LC/MS and immunoaffinity extraction. Anal Chem 10-1-2001;73(19):4704-4710. 11605850
  47. Shao, Y., Harris, A., Wang, M., Zhang, H., Cordell, G. A., Bowman, M., and Lemmo, E. Triterpene glycosides from Cimicifuga racemosa. J Nat Prod 2000;63(7):905-910. 10924163
  48. Wang, H. K., Sakurai, N., Shih, C. Y., and Lee, K. H. LC/TIS-MS fingerprint profiling of Cimicifuga species and analysis of 23-Epi-26-deoxyactein in Cimicifuga racemosa commercial products. J Agric Food Chem 3-9-2005;53(5):1379-1386. 15740010
  49. Fugh-Berman, A. and Ernst, E. Herb-drug interactions: reand assessment of report reliability. Br J Clin Pharmacol 2001;52(5):587-595. 11736868
  50. Chen, S. N., Lankin, D. C., Nikolic, D., Fabricant, D. S., Lu, Z. Z., Ramirez, B., van Breemen, R. B., Fong, H. H., Farnsworth, N. R., and Pauli, G. F. Chlorination diversifies Cimicifuga racemosa triterpene glycosides. J Nat Prod 2007;70(6):1016-1023. 17555351
  51. He, K., Zheng, B., Kim, C. H., Rogers, L., and Zheng, Q. Direct analysis and identification of triterpene glycosides by LC/MS in black cohosh, Cimicifuga racemosa, and in several commercially available black cohosh products. Planta Med 2000;66(7):635-640. 11105569
  52. Koeda, M., Aoki, Y., Sakurai, N., and Nagai, M. Studies on the Chinese crude drug "shoma." IX. Three novel cyclolanostanol xylosides, cimicifugosides H-1, H-2 and H-5, from cimicifuga rhizome. Chem Pharm Bull (Tokyo) 1995;43(5):771-776. 7553963
  53. Kennelly, E. J., Baggett, S., Nuntanakorn, P., Ososki, A. L., Mori, S. A., Duke, J., Coleton, M., and Kronenberg, F. Analysis of thirteen populations of black cohosh for formononetin. Phytomedicine 2002;9(5):461-467. 12222669
  54. Johnson, B. M. and van Breemen, R. B. In vitro formation of quinoid metabolites of the dietary supplement Cimicifuga racemosa (black cohosh). Chem Res Toxicol 2003;16(7):838-846. 12870886
  55. Wende, K., Mugge, C., Thurow, K., Schopke, T., and Lindequist, U. Actaeaepoxide 3-O-beta-D-xylopyranoside, a new cycloartane glycoside from the rhizomes of Actaea racemosa (Cimicifuga racemosa). J Nat Prod 2001;64(7):986-989. 11473444
  56. Chen, S. N., Fabricant, D. S., Pauli, G. F., Fong, H. H., and Farnsworth, N. R. Synthesis of cimiracemate B, a phenylpropanoid found in Cimicifuga racemosa. Nat Prod Res 2005;19(3):287-290. 15702643
  57. Hostanska, K., Nisslein, T., Freudenstein, J., Reichling, J., and Saller, R. Evaluation of cell death caused by triterpene glycosides and phenolic substances from Cimicifuga racemosa extract in human MCF-7 breast cancer cells. Biol Pharm Bull 2004;27(12):1970-1975. 15577215
  58. Li, W., Sun, Y., Liang, W., Fitzloff, J. F., and van Breemen, R. B. Identification of caffeic acid derivatives in Actea racemosa (Cimicifuga racemosa, black cohosh) by liquid chromatography/tandem mass spectrometry. Rapid Commun Mass Spectrom 2003;17(9):978-982. 12717772
  59. Nuntanakorn, P., Jiang, B., Yang, H., Cervantes-Cervantes, M., Kronenberg, F., and Kennelly, E. J. Analysis of polyphenolic compounds and radical scavenging activity of four American Actaea species. Phytochem Anal 2007;18(3):219-228. 17500365
  60. Hamburger, M. New approaches in analyzing the pharmacological properties of herbal extracts. Proc West Pharmacol Soc 2007;50:156-161. 18605255
  61. Rhyu, M. R., Lu, J., Webster, D. E., Fabricant, D. S., Farnsworth, N. R., and Wang, Z. J. Black cohosh (Actaea racemosa, Cimicifuga racemosa) behaves as a mixed competitive ligand and partial agonist at the human mu opiate receptor. J Agric Food Chem 12-27-2006;54(26):9852-9857. 17177511
  62. Einbond, L. S., Shimizu, M., Ma, H., Wu, H. A., Goldsberry, S., Sicular, S., Panjikaran, M., Genovese, G., and Cruz, E. Actein inhibits the Na+-K+-ATPase and enhances the growth inhibitory effect of digitoxin on human breast cancer cells. Biochem Biophys Res Commun 10-31-2008;375(4):608-613. 18755149
  63. Hostanska, K., Nisslein, T., Freudenstein, J., Reichling, J., and Saller, R. Inhibitory effect of an isopropanolic extract of black cohosh on the invasiveness of MDA-mB 231 human breast cancer cells. In Vivo<.em> 2007;21(2):349-355. 17436587
  64. Einbond, L. S., Su, T., Wu, H. A., Friedman, R., Wang, X., Ramirez, A., Kronenberg, F., and Weinstein, I. B. The growth inhibitory effect of actein on human breast cancer cells is associated with activation of stress response pathways. Int J Cancer 11-1-2007;121(9):2073-2083. 17621630
  65. Einbond, L. S., Su, T., Wu, H. A., Friedman, R., Wang, X., Jiang, B., Hagan, T., Kennelly, E. J., Kronenberg, F., and Weinstein, I. B. Gene expression analysis of the mechanisms whereby black cohosh inhibits human breast cancer cell growth. Anticancer Res 2007;27(2):697-712. 17465192
  66. Beck, V., Unterrieder, E., Krenn, L., Kubelka, W., and Jungbauer, A. Comparison of hormonal activity (estrogen, androgen and progestin) of standardized plant extracts for large scale use in hormone replacement therapy. J Steroid Biochem Mol Biol 2003;84(2-3):259-268. 12711012
  67. Nisslein, T. and Freudenstein, J. Concomitant administration of an isopropanolic extract of black cohosh and tamoxifen in the in vivo tumor model of implanted RUCA-I rat endometrial adenocarcinoma cells. Toxicol Lett 5-2-2004;150(3):271-275. 15110078
  68. Oerter, Klein K., Janfaza, M., Wong, J. A., and Chang, R. J. Estrogen bioactivity in fo-ti and other herbs used for their estrogen-like effects as determined by a recombinant cell bioassay. J Clin Endocrinol Metab 2003;88(9):4077-4079. 12970265
  69. Bodinet, C. and Freudenstein, J. Influence of marketed herbal menopause preparations on MCF-7 cell proliferation. Menopause 2004;11(3):281-289. 15167307
  70. Garita-Hernandez, M., Calzado, M. A., Caballero, F. J., Macho, A., Munoz, E., Meier, B., Brattstrom, A., Fiebich, B. L., and Appel, K. The growth inhibitory activity of the Cimicifuga racemosa extract Ze 450 is mediated through estrogen and progesterone receptors-independent pathways. Planta Med 2006;72(4):317-323. 16557472
  71. Stute, P., Nisslein, T., Gotte, M., Kamischke, A., Kiesel, L., and Klockenbusch, W. Effects of black cohosh on estrogen biosynthesis in normal breast tissue in vitro. Maturitas 8-20-2007;57(4):382-391. 17548177
  72. Reame, N. E., Lukacs, J. L., Padmanabhan, V., Eyvazzadeh, A. D., Smith, Y. R., and Zubieta, J. K. Black cohosh has central opioid activity in postmenopausal women: evidence from naloxone blockade and positron emission tomography neuroimaging. Menopause 2008;15(5):832-840. 18521048
  73. Hemmi, H., Kusano, G., and Ishida, N. Selective inhibition of nucleoside transport into mouse lymphoma L- 5178Y cells by cimicfugoside. J Pharmacobiodyn 1980;3(12):636-642. 7277178
  74. Hemmi H, Kitame F, Ishida N, and et al. Inhibition of thymidine transport into phytohemagglutinin-stimulated lymphocytes by triterpenoids from Cimicifuga species. J Pharm Dyn 1979;2:339-349.
  75. Gurley, B. J., Barone, G. W., Williams, D. K., Carrier, J., Breen, P., Yates, C. R., Song, P. F., Hubbard, M. A., Tong, Y., and Cheboyina, S. Effect of milk thistle (Silybum marianum) and black cohosh (Cimicifuga racemosa) supplementation on digoxin pharmacokinetics in humans. Drug Metab Dispos 2006;34(1):69-74. 16221754
Licensed by Natural Standard Copyright © 2010 by Natural Standard Corporation. All Rights Reserved.
back to Plant Profiler