Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Nutrition Research > Learning Center > Plant Profiler > Turmeric, Curcumin (Curcuma longa)
Plant Profiler
Turmeric, Curcumin (Curcuma longa)
Synonyms / Common Names / Related Terms
Amomoum curcuma, anlatone (constituent), ar-tumerone, CUR, Curcuma, Curcuma aromatica, Curcuma aromatica Salisbury, Curcuma domestica, Curcuma domestica Valet, Curcuma longa, Curcuma longa Linn., Curcuma longa rhizoma, curcuma oil, curcumin, diferuloylmethane, E zhu, Gelbwurzel, gurkemeje, Haldi, Haridra, Indian saffron, Indian yellow root, Jiang huang, kunir, kunyit, Kurkumawurzelstock, Kyoo, Number Ten (NT), Olena, Radix zedoaria longa, Rhizome de curcuma, safran des Indes, sesquiterpenoids, shati, tumeric, turmeric oil, turmeric root, tumerone (constituent), Ukon, yellowroot, Zedoary, Zingiberaceae (family), zingiberene, Zitterwurzel.
Mechanism of Action
Pharmacology:
  • Constituents: Curcumin (diferuloylmethane), a polyphenol compound responsible for the bright yellow color of turmeric, is believed to be the principal pharmacological agent. It is prepared from the roots of Curcuma longa.66 In addition to curcumin, turmeric contains the curcuminoids atlantone, bisdemethoxycurcumin, demethoxycurcumin, diaryl heptanoids, and tumerone. Turmeric also contains sesquiterpenoids7 and the constituent ar-tumerone67. Other constituents include sugars, resins, proteins, vitamins, and minerals (including iron and potassium).
  • Alzheimer's effects: Beta-Amyloid (betaA)-induced oxidative stress is a well-established pathway of neuronal cell death in Alzheimer's disease.2 Three curcuminoids from turmeric (Curcuma longa L.), including curcumin, demethoxycurcumin, and bisdemethoxycurcumin, were found to protect PC12 rat pheochromocytoma and normal human umbilical vein endothelial (HUVEC) cells from betaA(1-42) insult. These compounds may protect the cells from betaA(1-42) insult through antioxidant pathways. Other animal studies of Alzheimer's disease also suggest that curcumin may reduce levels of amyloid and oxidized proteins and prevent cognitive deficits.1 One alternative mechanism of action for these effects suggested by Baum et al. is metal chelation, which may reduce amyloid aggregation or oxidative neurotoxicity. Since curcumin more readily binds the redox-active metals and than the redox-inactive , curcumin might exert a net protective effect against beta toxicity or might suppress inflammatory damage by preventing metal induction of NF-kappaB. Mouse studies that evaluated the effects of dietary curcumin on inflammation, oxidative damage, and plaque pathology demonstrated that both low and high doses of curcumin significantly lowered oxidized proteins and interleukin-1beta, which is a proinflammatory cytokine elevated in the brains of these mice.3 Low-dose but not high-dose curcumin treatment has been shown to reduce the astrocytic marker GFAP and significantly decrease insoluble beta-amyloid (Abeta), soluble Abeta, and plaque burden by 43-50%. However, levels of amyloid precursor (APP) in the membrane fraction were not reduced.
  • Antibacterial effects: The ethyl acetate extract of Curcuma longa L. has demonstrated a higher antibacterial activity than the methanol extract or water extract.4
  • Anti-inflammatory effects: Turmeric has been associated with the inhibition of tumor necrosis factor-α, interleukin-8, monocyte inflammatory protein-1, interleukin-1B, and monocyte chemotactic protein-1 25. Turmeric and its constituent curcumin have been found to inhibit lipoxygenase and cyclooxygenase in rat tissues and in vitro23,18,22, as well as thromboxane B219 and leukotriene B4 formation28,23. Based on animal study, oral administration of curcumin may reduce expression of several cytokines, chemokines, and proteinases known to mediate aneurismal degeneration.36 In rat macrophages, curcumin inhibits the incorporation of arachidonic acid into membrane lipids, as well as prostaglandin E2, leukotriene B4, and leukotriene C4, but does not affect the release of arachidonic acid.30 Curcumin also inhibits the secretion of collagenase, elastase, and hyaluronidase. Inhibition of neutrophil function has been noted24, and in vitro research demonstrates that curcumin inhibits 5-hydroxy-eicosatetraenoic acid (5-HETE) in intact human neutrophils.18 Turmeric has been found to block cytokine-induced transcription of leukocyte adhesion molecules ICAM-1, VCAM-1, and E-selectin29, and it appears to induce the production of endogenous TGF-B1 in animal wounds33. Curcumin down-regulates transcription of genes responsible for the production of chemotactic cytokines in bone marrow stromal cells.35 Curcumin reduces chemically-induced rat paw edema32,31 and liver inflammation34.
  • Antioxidant effects: Turmeric has been reported to possess antioxidant properties in vitro and in animal studies.59,60,61,62,63,11,64,65,35 Turmeric preparations have been found to scavenge free radicals (peroxides) and phenolic oxidants29, inhibit lipid peroxidation induced by chemical agents68,69, and inhibit iron-dependent lipid peroxidation in rat tissues.70,71 In vitro research shows that turmeric may prevent oxidative damage to DNA72 and may be a potent scavenger of nitric acid65. Curcumin appears to generate a hydroxyl radical73. Structural features of curcuminoids that may contribute to antioxidant activity include phenolic and methoxy groups on phenyl rings and diketones.74 Research using aqueous extracts of turmeric suggests that curcumin is not the only antioxidant in turmeric 75, and turmerin has been identified as a water-soluble peptide from turmeric with antioxidant properties 76. Animal studies have reported the reversal of hepatonecrosis and fatty changes associated with turmeric, with reversal of aflatoxin-induced liver damage 8.
  • Anti-platelet aggregation effects: Curcumin inhibits thromboxane A2 without affecting the synthesis of prostaglandin I220 In vitro, curcumin inhibits platelet aggregation induced by ADP, epinephrine, or collagen.21,19 Turmeric appears to inhibit arachidonic acid incorporation into platelet phospholipids, degradation of phospholipids, and cyclooxygenase.77,9
  • Anti-proliferative effects: Multiple pre-clinical studies have explored potential anti-cancer mechanisms of curcumin.37,39,40,41,42,43,44,45,46,5,47,48,49,6,15,12,51,52,53,54,55,56 In a rat model, the effects of 0.2% or 0.6% dietary curcumin were evaluated on chemically induced colon adenocarcinoma.50 Histological examination after one year revealed both preventative and therapeutic benefits of curcumin when compared to animals not receiving curcumin, with better response at higher doses. Histologic examination revealed evidence of apoptosis of cancer cells. In mice, six weeks of a 2% curcumin diet was found to decrease cellular proliferation and increase apoptosis of implanted androgen-dependent LNCaP prostate cancer cells.45 However, in vitro research on the anti-proliferative effects of curcumin on breast cancer cells reported no evidence of apoptosis.78 Dietary turmeric extract given to mice (2% or 5% of diet) significantly inhibited chemically-induced skin and gastric tumors.38 A 57% reduction in the incidence of chemically-induced colonic epithelial cell dysplasia was noted in mice fed a 2% curcumin diet.79 Curcumin has been found to inhibit the formation of chemically-induced aberrant crypt foci in rat colons, and to inhibit colonic mucosal tyrosine protein kinase activity.22 In vitro, curcumin derivatives demonstrated anti-tumor effects against leukemic cells, and in test animals, they decreased the incidence of experimentally-induced forestomach tumors by 20% and the number of skin tumors and papillomas by 68% and 57%, respectively.52 HL-60 leukemia cells are susceptible to curcumin treatment in vitro.80 Curcumin may suppress telomerase activity and induce apoptosis in human cancer cell lines.40 Extracts of turmeric and curcumin have been found to be to be non-mutagenic using the Ames test and to serve protective roles against experimental mutagenesis induced by capsaicin, chili extract, and tobacco-derived mutagens.14 In a different animal study, however, concentrations up to 5mM curcumin did not protect against cytotoxicity of paracetamol in freshly isolated hepatocytes, and curcumin itself was found to be mildly hepatotoxic at high doses.13 Curcumin inhibits the growth of human epidermoid carcinoma A431 cells in vitro.81 Topical application of curcumin inhibits chemically-induced tumor promotion in mouse skin82, which may be attributable to suppression of protein kinase C activity83. Curcumins have also been noted in vitro to inhibit the nitrosation of methyl urea by sodium nitrate.84 In vitro research suggests that curcumin exerts activity against human breast cancer cells56, including hormone-dependent, hormone-independent, and multidrug-resistant cell lines78, which may occur via effects on aryl hydrocarbon receptors57 or ornithine decarboxylase activity78. The protective role of curcumin against carcinogenesis has also been attributed to antioxidant effects.72 In rats, curcumin is reported to be protective against radiation-induced lung fibrosis and micronuclei formation85 and to protect against acute adriamycin-induced myocardial toxicity 58. In rat aortic smooth muscle cells, curcumin inhibited cell proliferation, arrested cell cycle progression, and induced cell apoptosis.86 In vitro, curcumin has exhibited activity against human B-cells immortalized by Epstein-Barr virus.87 Anti-angiogenic properties have been proposed.88,89,45,90
  • Lipid-lowering effects: In rat models of hyperlipidemia, a diet of 0.5% curcumin for eight weeks significantly lowered serum low-density lipoprotein (LDL), very low-density lipoprotein (VLDL), total cholesterol, and triglyceride levels, possibly by enhancing the activity of hepatic cholesterol-7a-hydroxylase and increasing cholesterol catabolism.91,17,16 The turmeric constituents demethoxycurcumin, bisdemethoxycurcumin, and acetylcurcumin appear to inhibit -stimulated lipid peroxidation in rat tissues and liver microsomes.69 In a rat model of hyperlipidemia, a 50% ethanolic extract of turmeric was associated with a significant reduction in the ratio of total cholesterol to phospholipids.92 In rabbits fed a high cholesterol diet, oral turmeric (1.6-3.2mg/kg) was associated with lower levels of plasma cholesterol and triglycerides than a control group, although no differences in atherogenesis were noted on histological examination of aortas.93 Cholesterol levels were lower in the 1.6mg/kg group.
  • Gastro-protective effects: Oral administration of turmeric to rats (500mg/kg) significantly reduces the incidence of chemically-induced duodenal ulcers and is associated with an increase in intestinal wall mucus and non-protein sulfhydryl content.26 However, early research in guinea pigs reported that various constituents of turmeric do not protect against histamine-induced gastric ulcerations.94
  • Gallbladder effects: Gallbladder contraction over the two-hour period following the administration of 20mg curcumin has been demonstrated in humans.10 Animal research reports that curcumin in the diet reduces the incidence of chemically-induced gallstones in mice.95
  • Hypoglycemic effects: Based on animal study, both curcuminoids and sesquiterpenoids in turmeric may exhibit hypoglycemic effects via PPAR-gamma activation.7
  • Weight loss effects: In a rat study that investigated a Chinese herbal formulation called Number Ten for weight loss, leptin and body fat decreased significantly in the treatment group compared to the control group (p<0.009 and p<0.006, respectively).
  • Other effects: Chelation: In vitro research on liposomal membranes has demonstrated that curcumin forms chelates with iron.96,97 Phototoxicity: Curcumin in low concentrations has been found to potentiate phototoxicity to the bacteria S. typhimurium and E. coli.98
Pharmacodynamics/Kinetics:
  • Absorption: Animal research shows that the absorption of curcumin after oral administration varies from 25-60%, with most of the absorbed flavonoid being metabolized in the intestinal mucosa and liver.99 The remainder is excreted in the feces.100
  • Distribution: Based on a clinical trial, Garcea et al. report that a daily dose of 3.6g curcumin may achieve pharmacologically efficacious levels in the colorectum with negligible distribution of curcumin outside the gut.5
  • Bioavailability: Authors of one clinical trial concluded that the lack of quantifiable curcumin in the plasma observed after a dose as high as 3,600mg is consistent with recent clinical reports, in which oral doses of 30-180mg curcumin failed to establish detectable plasma levels, and doses of 4,000-12,000mg yielded curcumin peak levels of 0.5-2mcM/L after one hour of administration.5 The authors also concluded that products of metabolic curcumin conjugation are present in the colorectum of humans who have ingested curcumin, but that these metabolites only contribute to a very minor extent of the overall colorectal load of curcumin-derived species. This finding is consistent with the idea that the pharmacologic effects of curcumin in the colorectum are probably caused by the parent compound and not by its metabolites.5
  • Pharmacodynamics: In rats, curcumin is reported to be a potent inhibitor of cytochrome P450 (CYP) 1A1/1A2, a less potent inhibitor of CYP 2B1/2B2, and a weak inhibitor of CYP 2E1.27 Inhibition of cytochrome P450 has also been demonstrated in vitro .52 Turmeric may decrease hepatocyte glutathione levels101; curcumin appears to induce glutathion-s-transferase activity in mice102.
  • Curcumin, a constituent of turmeric, completely inhibited mycelial growth of Aspergillus alliaceus isolate 791 at 0.1% (w/v) and decreased ochratoxin A production by approximately 70% at 0.01% (w/v) .103
  • In the checkerboard test, the ethyl acetate extract of Curcuma longa L. markedly lowered the MICs of ampicillin and oxacillin against methicillin-resistant Staphylococcus aureus (MRSA).4 In the bacterial invasion assay, MRSA intracellular invasion was significantly decreased in the presence of 0.125-2mg/mL of Curcuma longa extract compared to the control group.
References
  1. Baum, L. and Ng, A. Curcumin interaction with copper and iron suggests one possible mechanism of action in Alzheimer's disease animal models. J Alzheimers Dis 2004;6(4):367-377. 15345806
  2. Kim, D. S., Park, S. Y., and Kim, J. K. Curcuminoids from Curcuma longa L. (Zingiberaceae) that protect PC12 rat pheochromocytoma and normal human umbilical vein endothelial cells from betaA(1-42) insult. Neurosci Lett 4-27-2001;303(1):57-61. 11297823
  3. Lim, G. P., Chu, T., Yang, F., Beech, W., Frautschy, S. A., and Cole, G. M. The curry spice curcumin reduces oxidative damage and amyloid pathology in an Alzheimer transgenic mouse. J Neurosci 11-1-2001;21(21):8370-8377. 11606625
  4. Kim, K. J., Yu, H. H., Cha, J. D., Seo, S. J., Choi, N. Y., and You, Y. O. Antibacterial activity of Curcuma longa L. against methicillin-resistant Staphylococcus aureus. Phytother Res 2005;19(7):599-604. 16161063
  5. Garcea, G., Berry, D. P., Jones, D. J., Singh, R., Dennison, A. R., Farmer, P. B., Sharma, R. A., Steward, W. P., and Gescher, A. J. Consumption of the putative chemopreventive agent curcumin by cancer patients: assessment of curcumin levels in the colorectum and their pharmacodynamic consequences. Cancer Epidemiol Biomarkers Prev 2005;14(1):120-125. 15668484
  6. Joshi, J., Ghaisas, S., Vaidya, A., Vaidya, R., Kamat, D. V., Bhagwat, A. N., and Bhide, S. Early human safety study of turmeric oil (Curcuma longa oil) administered orally in healthy volunteers. J Assoc Physicians India 2003;51:1055-1060. 15260388
  7. Nishiyama, T., Mae, T., Kishida, H., Tsukagawa, M., Mimaki, Y., Kuroda, M., Sashida, Y., Takahashi, K., Kawada, T., Nakagawa, K., and Kitahara, M. Curcuminoids and sesquiterpenoids in turmeric (Curcuma longa L.) suppress an increase in blood glucose level in type 2 diabetic KK-Ay mice. J Agric Food Chem 2-23-2005;53(4):959-963. 15713005
  8. Soni, K. B., Rajan, A., and Kuttan, R. Reversal of aflatoxin induced liver damage by turmeric and curcumin. Cancer Lett 9-30-1992;66(2):115-121. 1394115
  9. Shah, B. H., Nawaz, Z., Pertani, S. A., Roomi, A., Mahmood, H., Saeed, S. A., and Gilani, A. H. Inhibitory effect of curcumin, a food spice from turmeric, on platelet- activating factor- and arachidonic acid-mediated platelet aggregation through inhibition of thromboxane formation and Ca2+ signaling. Biochem Pharmacol 10-1-1999;58(7):1167-1172. 10484074
  10. Rasyid A and Lelo A. The effect of curcumin and placebo on human gall-bladder function: an ultrasound study. Aliment Pharmacol Ther 1999;13(2):245-249.
  11. Ramirez-Bosca A, Soler A, Guiterrez MA, and et al. Antioxidant curcuma extracts decrease the blood lipid peroxide levels of human subjects. Age 1995;18(4):167-169.
  12. Kuttan, R., Sudheeran, P. C., and Josph, C. D. Turmeric and curcumin as topical agents in cancer therapy. Tumori 2-28-1987;73(1):29-31. 2435036
  13. Donatus, I. A., Sardjoko, and Vermeulen, N. P. Cytotoxic and cytoprotective activities of curcumin. Effects on paracetamol-induced cytotoxicity, lipid peroxidation and glutathione depletion in rat hepatocytes. Biochem Pharmacol  6-15-1990;39(12):1869-1875. 2353930 
  14. Nagabhushan, M., Amonkar, A. J., and Bhide, S. V. In vitro antimutagenicity of curcumin against environmental mutagens. Food Chem Toxicol 1987;25(7):545-547. 3623345
  15. Kuttan, R., Bhanumathy, P., Nirmala, K., and George, M. C. Potential anticancer activity of turmeric (Curcuma longa). Cancer Lett 1985;29(2):197-202. 4075289
  16. Babu, P. S. and Srinivasan, K. Hypolipidemic action of curcumin, the active principle of turmeric (Curcuma longa) in streptozotocin induced diabetic rats. Mol Cell Biochem 1997;166(1-2):169-175. 9046034
  17. Srinivasan K and Sambaiah K. The effect of spices on cholesterol 7 alpha-hydroxylase activity and on serum and hepatic cholesterol levels in the rat. Internat J Vit Nutr Res 1991;61:364-369.
  18. Flynn, D. L., Rafferty, M. F., and Boctor, A. M. Inhibition of 5-hydroxy-eicosatetraenoic acid (5-HETE) formation in intact human neutrophils by naturally-occurring diarylheptanoids: inhibitory activities of curcuminoids and yakuchinones. Prostaglandins Leukot Med  1986;22(3):357-360. 3460103
  19. Srivastava, K. C., Bordia, A., and Verma, S. K. Curcumin, a major component of food spice turmeric (Curcuma longa) inhibits aggregation and alters eicosanoid metabolism in human blood platelets. Prostaglandins Leukot Essent Fatty Acids 1995;52(4):223-227. 7784468
  20. Srivastava, R., Dikshit, M., Srimal, R. C., and Dhawan, B. N. Anti-thrombotic effect of curcumin. Thromb Res 11-1-1985;40(3):413-417. 4082116
  21. Srivastava, R., Puri, V., Srimal, R. C., and Dhawan, B. N. Effect of curcumin on platelet aggregation and vascular prostacyclin synthesis. Arzneimittelforschung. 1986;36(4):715-717. 3521617
  22. Rao, C. V., Simi, B., and Reddy, B. S. Inhibition by dietary curcumin of azoxymethane-induced ornithine decarboxylase, tyrosine protein kinase, arachidonic acid metabolism and aberrant crypt foci formation in the rat colon. Carcinogenesis 1993;14(11):2219-2225. 8242846
  23. Ammon, H. P., Safayhi, H., Mack, T., and Sabieraj, J. Mechanism of antiinflammatory actions of curcumine and boswellic acids. J Ethnopharmacol 1993;38(2-3):113-119. 8510458
  24. Srivastava, R. Inhibition of neutrophil response by curcumin. Agents Actions 1989;28(3-4):298-303. 2556903
  25. Abe, Y., Hashimoto, S., and Horie, T. Curcumin inhibition of inflammatory cytokine production by human peripheral blood monocytes and alveolar macrophages. Pharmacol Res 1999;39(1):41-47. 10051376
  26. Rafatullah, S., Tariq, M., Al Yahya, M. A., Mossa, J. S., and Ageel, A. M. Evaluation of turmeric (Curcuma longa) for gastric and duodenal antiulcer activity in rats. J Ethnopharmacol 1990;29(1):25-34. 2345457
  27. Oetari, S., Sudibyo, M., Commandeur, J. N., Samhoedi, R., and Vermeulen, N. P. Effects of curcumin on cytochrome P450 and glutathione S-transferase activities in rat liver. Biochem Pharmacol 1-12-1996;51(1):39-45. 8534266
  28. Ammon, H. P., Anazodo, M. I., Safayhi, H., Dhawan, B. N., and Srimal, R. C. Curcumin: a potent inhibitor of leukotriene B4 formation in rat peritoneal polymorphonuclear neutrophils (PMNL). Planta Med 1992;58(2):226. 1326775
  29. Gupta, B. and Ghosh, B. Curcuma longa inhibits TNF-alpha induced expression of adhesion molecules on human umbilical vein endothelial cells. Int J Immunopharmacol 1999;21(11):745-757. 10576620
  30. Joe, B. and Lokesh, B. R. Effect of curcumin and capsaicin on arachidonic acid metabolism and lysosomal enzyme secretion by rat peritoneal macrophages. Lipids 1997;32(11):1173-1180. 9397403
  31. Mukhopadhyay, A., Basu, N., Ghatak, N., and Gujral, P. K. Anti-inflammatory and irritant activities of curcumin analogues in rats. Agents Actions 1982;12(4):508-515. 7180736
  32. Nurfina AN, Reksohadiprodjo MS, Timmerman H, and et al. Synthesis of some symmetrical curcumin derivatives and their antiinflammatory activity. Eur J Med Chem 1997;32:321-328.
  33. Sidhu, G. S., Singh, A. K., Thaloor, D., Banaudha, K. K., Patnaik, G. K., Srimal, R. C., and Maheshwari, R. K. Enhancement of wound healing by curcumin in animals. Wound Repair Regen 1998;6(2):167-177. 9776860
  34. Srimal RC, Khanna NM, and Dhawan BN. A preliminary report on anti-inflammatory activity of curcumin. Indian J Pharmacol 1971;3:10.
  35. Xu, Y. X., Pindolia, K. R., Janakiraman, N., Noth, C. J., Chapman, R. A., and Gautam, S. C. Curcumin, a compound with anti-inflammatory and anti-oxidant properties, down-regulates chemokine expression in bone marrow stromal cells. Exp Hematol 1997;25(5):413-422. 9168063
  36. Parodi, F. E., Mao, D., Ennis, T. L., Pagano, M. B., and Thompson, R. W. Oral administration of diferuloylmethane (curcumin) suppresses proinflammatory cytokines and destructive connective tissue remodeling in experimental abdominal aortic aneurysms. Ann Vasc Surg 2006;20(3):360-368. 16779518
  37. Aggarwal, B. B., Kumar, A., and Bharti, A. C. Anticancer potential of curcumin: preclinical and clinical studies. Anticancer Res 2003;23(1A):363-398. 12680238
  38. Azuine, M. A. and Bhide, S. V. Chemopreventive effect of turmeric against stomach and skin tumors induced by chemical carcinogens in Swiss mice. Nutr Cancer 1992;17(1):77-83. 1574446
  39. Bharti, A. C., Donato, N., Singh, S., and Aggarwal, B. B. Curcumin (diferuloylmethane) down-regulates the constitutive activation of nuclear factor-kappa B and IkappaBalpha kinase in human multiple myeloma cells, leading to suppression of proliferation and induction of apoptosis. Blood 2-1-2003;101(3):1053-1062. 12393461
  40. Cui, S. X., Qu, X. J., Xie, Y. Y., Zhou, L., Nakata, M., Makuuchi, M., and Tang, W. Curcumin inhibits telomerase activity in human cancer cell lines. Int J Mol.Med 2006;18(2):227-231. 16820928
  41. Chaudhary, L. R. and Hruska, K. A. Inhibition of cell survival signal protein kinase B/Akt by curcumin in human prostate cancer cells. J Cell Biochem 5-1-2003;89(1):1-5. 12682902
  42. Deeb, D., Xu, Y. X., Jiang, H., Gao, X., Janakiraman, N., Chapman, R. A., and Gautam, S. C. Curcumin (Diferuloyl-Methane) Enhances Tumor Necrosis Factor-related Apoptosis-inducing Ligand-induced Apoptosis in LNCaP Prostate Cancer Cells Mol Cancer Ther 2003;2(1):95-103. 12533677
  43. Devasena, T., Rajasekaran, K. N., Gunasekaran, G., Viswanathan, P., and Menon, V. P. Anticarcinogenic effect of bis-1,7-(2-hydroxyphenyl)-hepta-1,6-diene-3,5-dione a curcumin analog on DMH-induced colon cancer model. Pharmacol Res 2003;47(2):133-140. 12543061
  44. Di, G. H., Li, H. C., Shen, Z. Z., and Shao, Z. M. [Analysis of anti-proliferation of curcumin on human breast cancer cells and its mechanism]. Zhonghua Yi Xue Za Zhi 10-25-2003;83(20):1764-1768. 14642080
  45. Dorai, T., Cao, Y. C., Dorai, B., Buttyan, R., and Katz, A. E. Therapeutic potential of curcumin in human prostate cancer. III. Curcumin inhibits proliferation, induces apoptosis, and inhibits angiogenesis of LNCaP prostate cancer cells in vivo. Prostate 6-1-2001;47(4):293-303. 11398177
  46. Elattar, T. M. and Virji, A. S. The inhibitory effect of curcumin, genistein, quercetin and cisplatin on the growth of oral cancer cells in vitro. Anticancer Res 2000;20(3A):1733-1738. 10928101
  47. Gautam, S. C., Xu, Y. X., Pindolia, K. R., Janakiraman, N., and Chapman, R. A. Nonselective inhibition of proliferation of transformed and nontransformed cells by the anticancer agent curcumin (diferuloylmethane). Biochem Pharmacol 4-15-1998;55(8):1333-1337. 9719490
  48. Hastak, K., Lubri, N., Jakhi, S. D., More, C., John, A., Ghaisas, S. D., and Bhide, S. V. Effect of turmeric oil and turmeric oleoresin on cytogenetic damage in patients suffering from oral submucous fibrosis. Cancer Lett 6-24-1997;116(2):265-269. 9215873
  49. Jiang, M. C., Yang-Yen, H. F., Yen, J. J., and Lin, J. K. Curcumin induces apoptosis in immortalized NIH 3T3 and malignant cancer cell lines. Nutr Cancer 1996;26(1):111-120. 8844727
  50. Kawamori, T., Lubet, R., Steele, V. E., Kelloff, G. J., Kaskey, R. B., Rao, C. V., and Reddy, B. S. Chemopreventive effect of curcumin, a naturally occurring anti- inflammatory agent, during the promotion/progression stages of colon cancer. Cancer Res 2-1-1999;59(3):597-601. 9973206
  51. Moragoda, L., Jaszewski, R., and Majumdar, A. P. Curcumin induced modulation of cell cycle and apoptosis in gastric and colon cancer cells. Anticancer Res 2001;21(2A):873-878. 11396178
  52. Nagabhushan M and Bhide SV. Curcumin as an inhibitor of cancer. J Amer Coll Nutrit 1992;11(2):192-198.
  53. Pereira, M. A., Grubbs, C. J., Barnes, L. H., Li, H., Olson, G. R., Eto, I., Juliana, M., Whitaker, L. M., Kelloff, G. J., Steele, V. E., and Lubet, R. A. Effects of the phytochemicals, curcumin and quercetin, upon azoxymethane-induced colon cancer and 7,12-dimethylbenz[a]anthracene- induced mammary cancer in rats. Carcinogenesis 1996;17(6):1305-1311. 8681447
  54. Rashmi, R., Santhosh Kumar, T. R., and Karunagaran, D. Human colon cancer cells differ in their sensitivity to curcumin-induced apoptosis and heat shock protects them by inhibiting the release of apoptosis-inducing factor and caspases. FEBS Lett 3-13-2003;538(1-3):19-24. 12633846
  55. Sharma, R. A., McLelland, H. R., Hill, K. A., Ireson, C. R., Euden, S. A., Manson, M. M., Pirmohamed, M., Marnett, L. J., Gescher, A. J., and Steward, W. P. Pharmacodynamic and pharmacokinetic study of oral Curcuma extract in patients with colorectal cancer. Clin Cancer Res 2001;7(7):1894-1900. 11448902
  56. Verma S, Salamone E, and Goldin B. Curcumin and genistein, plant natural products, show synergistic inhibitory effects on the growth of human breast cancer MCF-7 cells induced by estrogenic pesticides. Biochem Biophys Res Commun 4-28-1997;233(3):692-696. 9168916
  57. Ciolino, H. P., Daschner, P. J., Wang, T. T., and Yeh, G. C. Effect of curcumin on the aryl hydrocarbon receptor and cytochrome P450 1A1 in MCF-7 human breast carcinoma cells. Biochem Pharmacol 7-15-1998;56(2):197-206. 9698073
  58. Venkatesan, N. Curcumin attenuation of acute adriamycin myocardial toxicity in rats. Br J Pharmacol 1998;124(3):425-427. 9647462
  59. Balogun, E., Hoque, M., Gong, P., Killeen, E., Green, C. J., Foresti, R., Alam, J., and Motterlini, R. Curcumin activates the haem oxygenase-1 gene via regulation of Nrf2 and the antioxidant-responsive element. Biochem J 5-1-2003;371(Pt 3):887-895. 12570874
  60. Braga, M. E., Leal, P. F., Carvalho, J. E., and Meireles, M. A. Comparison of yield, composition, and antioxidant activity of turmeric (Curcuma longa L.) extracts obtained using various techniques. J Agric Food Chem 10-22-2003;51(22):6604-6611. 14558784
  61. Iqbal, M., Sharma, S. D., Okazaki, Y., Fujisawa, M., and Okada, S. Dietary supplementation of curcumin enhances antioxidant and phase II metabolizing enzymes in ddY male mice: possible role in protection against chemical carcinogenesis and toxicity. Pharmacol Toxicol 2003;92(1):33-38. 12710595
  62. Motterlini, R., Foresti, R., Bassi, R., and Green, C. J. Curcumin, an antioxidant and anti-inflammatory agent, induces heme oxygenase-1 and protects endothelial cells against oxidative stress. Free Radic Biol Med 4-15-2000;28(8):1303-1312. 10889462
  63. Osawa, T., Sugiyama, Y., Inayoshi, M., and Kawakishi, S. Antioxidative activity of tetrahydrocurcuminoids. Biosci Biotechnol Biochem 1995;59(9):1609-1612. 8520105
  64. Ruby, A. J., Kuttan, G., Babu, K. D., Rajasekharan, K. N., and Kuttan, R. Anti-tumour and antioxidant activity of natural curcuminoids. Cancer Lett 7-20-1995;94(1):79-83. 7621448
  65. Sreejayan and Rao, M. N. Nitric oxide scavenging by curcuminoids. J Pharm Pharmacol 1997;49(1):105-107. 9120760
  66. Ali, R. E. and Rattan, S. I. Curcumin's biphasic hormetic response on proteasome activity and heat-shock protein synthesis in human keratinocytes. Ann N Y Acad Sci 2006;1067:394-399. 16804017
  67. Ji, M., Choi, J., Lee, J., and Lee, Y. Induction of apoptosis by ar-turmerone on various cell lines. Int J Mol Med 2004;14(2):253-256. 15254774
  68. Reddy, A. C. and Lokesh, B. R. Studies on the inhibitory effects of curcumin and eugenol on the formation of reactive oxygen species and the oxidation of ferrous iron. Mol Cell Biochem 8-17-1994;137(1):1-8. 7845373
  69. Sreejayan and Rao, M. N. Curcuminoids as potent inhibitors of lipid peroxidation. J Pharm Pharmacol 1994;46(12):1013-1016. 7714712
  70. Sreejayan and Rao MNA. Curcumin inhibits iron-dependent lipid peroxidation. Int J Pharmaceut 1993;100:93-97.
  71. Rao S and Rao M. Curcumin inhibits iron-dependent lipid peroxidation. Int J Pharmaceut 1993;1993:93-97.
  72. Subramanian, M., Sreejayan, Rao, M. N., Devasagayam, T. P., and Singh, B. B. Diminution of singlet oxygen-induced DNA damage by curcumin and related antioxidants. Mutat Res 12-1-1994;311(2):249-255. 7526190
  73. Kunchandy E and Rao MN. Effect of curcumin on hydroxyl radical generation through Fenton reaction. Int J Pharmaceut 1989;57:173-176.
  74. Sreejayan, N. and Rao, M. N. Free radical scavenging activity of curcuminoids. Arzneimittelforschung 1996;46(2):169-171. 8720307
  75. Shalini, V. K. and Srinivas, L. Lipid peroxide induced DNA damage: protection by turmeric (Curcuma longa). Mol Cell Biochem 1987;77(1):3-10. 3696162
  76. Srinivas, L. and Shalini, V. K. DNA damage by smoke: protection by turmeric and other inhibitors of ROS. Free Radic Biol Med 1991;11(3):277-283. 1937145
  77. Srivastava, K. C. Extracts from two frequently consumed spices--cumin (Cuminum cyminum) and turmeric (Curcuma longa)--inhibit platelet aggregation and alter eicosanoid biosynthesis in human blood platelets. Prostaglandins Leukot Essent Fatty Acids 1989;37(1):57-64. 2503839
  78. Mehta, K., Pantazis, P., McQueen, T., and Aggarwal, B. B. Antiproliferative effect of curcumin (diferuloylmethane) against human breast tumor cell lines. Anticancer Drugs 1997;8(5):470-481. 9215611
  79. Huang, M. T., Deschner, E. E., Newmark, H. L., Wang, Z. Y., Ferraro, T. A., and Conney, A. H. Effect of dietary curcumin and ascorbyl palmitate on azoxymethanol- induced colonic epithelial cell proliferation and focal areas of dysplasia. Cancer Lett 6-15-1992;64(2):117-121. 1611594
  80. Kuo, M. L., Huang, T. S., and Lin, J. K. Curcumin, an antioxidant and anti-tumor promoter, induces apoptosis in human leukemia cells. Biochim Biophys Acta 11-15-1996;1317(2):95-100. 8950193
  81. Korutla, L. and Kumar, R. Inhibitory effect of curcumin on epidermal growth factor receptor kinase activity in A431 cells. Biochim Biophys Acta 12-30-1994;1224(3):597-600. 7803521
  82. Conney, A. H., Lysz, T., Ferraro, T., Abidi, T. F., Manchand, P. S., Laskin, J. D., and Huang, M. T. Inhibitory effect of curcumin and some related dietary compounds on tumor promotion and arachidonic acid metabolism in mouse skin. Adv Enzyme Regul 1991;31:385-396. 1908616
  83. Liu, J. Y., Lin, S. J., and Lin, J. K. Inhibitory effects of curcumin on protein kinase C activity induced by 12-O-tetradecanoyl-phorbol-13-acetate in NIH 3T3 cells. Carcinogenesis 1993;14(5):857-861. 8504477
  84. Nagabhushan, M., Nair, U. J., Amonkar, A. J., D'Souza, A. V., and Bhide, S. V. Curcumins as inhibitors of nitrosation in vitro. Mutat Res 1988;202(1):163-169. 3054526
  85. Thresiamma, K. C., George, J., and Kuttan, R. Protective effect of curcumin, ellagic acid and bixin on radiation induced toxicity. Indian J Exp Biol 1996;34(9):845-847. 9014516
  86. Chen, H. W. and Huang, H. C. Effect of curcumin on cell cycle progression and apoptosis in vascular smooth muscle cells. Br J Pharmacol 1998;124(6):1029-1040. 9720770
  87. Ranjan, D., Siquijor, A., Johnston, T. D., Wu, G., and Nagabhuskahn, M. The effect of curcumin on human B-cell immortalization by Epstein-Barr virus. Am Surg 1998;64(1):47-51. 9457037
  88. Robinson, T. P., Ehlers, T., Hubbard IV, R. B., Bai, X., Arbiser, J. L., Goldsmith, D. J., and Bowen, J. P. Design, synthesis, and biological evaluation of angiogenesis inhibitors: aromatic enone and dienone analogues of curcumin. Bioorg Med Chem Lett 1-6-2003;13(1):115-117. 12467629
  89. Gao, C., Ding, Z., Liang, B., Chen, N., and Cheng, D. [Study on the effects of curcumin on angiogenesis]. Zhong Yao Cai 2003;26(7):499-502. 14650061
  90. Thaloor, D., Singh, A. K., Sidhu, G. S., Prasad, P. V., Kleinman, H. K., and Maheshwari, R. K. Inhibition of angiogenic differentiation of human umbilical vein endothelial cells by curcumin. Cell Growth Differ 1998;9(4):305-312. 9563850
  91. Babu, P. S. and Srinivasan, K. Influence of dietary curcumin and cholesterol on the progression of experimentally induced diabetes in albino rat. Mol Cell Biochem 11-8-1995;152(1):13-21. 8609907
  92. Dixit, V. P., Jain, P., and Joshi, S. C. Hypolipidaemic effects of Curcuma longa L and Nardostachys jatamansi, DC in triton-induced hyperlipidaemic rats. Indian J Physiol Pharmacol 1988;32(4):299-304. 3215683
  93. Ramirez-Tortosa, M. C., Mesa, M. D., Aguilera, M. C., Quiles, J. L., Baro, L., Ramirez-Tortosa, C. L., Martinez-Victoria, E., and Gil, A. Oral administration of a turmeric extract inhibits LDL oxidation and has hypocholesterolemic effects in rabbits with experimental atherosclerosis. Atherosclerosis 1999;147(2):371-378. 10559523
  94. Bhatia A, Singh GB, and Khanna NM. Effect of curcumin, its alkali salts & Curcuma longa oil in histamine-induced gastric ulceration. Indian J Exp Biol 1964;2:158-160.
  95. Hussain, M. S. and Chandrasekhara, N. Effect on curcumin on cholesterol gall-stone induction in mice. Indian J Med.Res 1992;96:288-291. 1459671
  96. Tonnesen HH. Chemistry of curcumin and curcuminoids. Phenol Comp Food Effects Health 1992;143-153.
  97. Tonnesen HH and Greenhill JV. Studies on curcumin and curcuminoids. XXII: Curcumin as a reducing agent and as a radical scavenger. Int J Pharmaceut 1992;87:79-87.
  98. Tonnesen, H. H., de Vries, H., Karlsen, J., and Beijersbergen, van Henegouwen. Studies on curcumin and curcuminoids. IX: Investigation of the photobiological activity of curcumin using bacterial indicator systems. J Pharm Sci 1987;76(5):371-373. 3309256
  99. Wahlstrom, B. and Blennow, G. A study on the fate of curcumin in the rat. Acta Pharmacol Toxicol (Copenh) 1978;43(2):86-92. 696348
  100. Ravindranath, V. and Chandrasekhara, N. Absorption and tissue distribution of curcumin in rats. Toxicology 1980;16(3):259-265. 7423534
  101. Mathews S and Rao MNA. Interaction of curcumin with glutathione. Int J Pharmaceut 1991;76:257-259.
  102. Susan, M. and Rao, M. N. Induction of glutathione S-transferase activity by curcumin in mice. Arzneimittelforschung 1992;42(7):962-964. 1418063
  103. Lee, S. E., Park, B. S., Bayman, P., Baker, J. L., Choi, W. S., and Campbell, B. C. Suppression of ochratoxin biosynthesis by naturally occurring alkaloids. Food Addit Contam 2007;24(4):391-397. 17454112
Licensed by Natural Standard Copyright © 2010 by Natural Standard Corporation. All Rights Reserved.
back to Plant Profiler
back to top