Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Nutrition Research > Learning Center > Plant Profiler > Lemongrass (Cymbopogon)
Plant Profiler
Lemongrass (Cymbopogon)
Lemongrass (Cymbopogon) Image
Synonyms / Common Names / Related Terms
Abafado (Portuguese), alpha-citral, alpha-terpineole, Andropogon citratus, Andropogon nardus, bai mak nao (Lao), beta-citral (neral), beta-myrcene, bhustrina (Indian), British Indian lemongrass, capim-cidrao, carene-2, caspase-3, Ceylon citronella grass, citral, Cochin lemongrass, Cymbopogon ambiguus, Cymbopogon citrates, Cymbopogon citratus DC, Cymbopogon excavatus, Cymbopogon flexuosus, Cymbopogon goeringii, Cymbopogon martinii, Cymbopogon nardus, Cymbopogon nardus, Cymbopogon proximus, Cymbopogon schoenanthus (L.), Cymbopogon winterianus, Cymbopogon winterianus Jowitt, East Indian lemongrass, erba di limone (Italian), essência de capim-limão (Portuguese), essential oils, farnesol, fever grass, geraniol , geranium grass, geranyl acetate, Graminaeae (family), Guatemala lemongrass, Halfa barr, herbe de citron (French), hierba de limon (Spanish), Java citronella, lemon grass, lemon grass extract (LGE), lemongrass oil, lemon grass oil (LGO), lemongrass stalk, lemon herbs, Lg, Madagascar lemongrass, Melissa grass, MYR, myrcene, palmarosa, pinene, piperitone, Poaceae (family), proximadiol, Santalum acuminatum, sera (Indian, Sinhalese), serai (Malay), sere (Indonesian), sereh (Indonesian), Sudanese flora, takrai (Thai), terpene beta-myrcene, West Indian lemongrass, Zitronengras (German).
Note: This review does not include citronella oil or stone root.
Mechanism of Action
Pharmacology:
  • Constituents: One of the main constituents of the many different species of lemongrass (genus Cymbopogon) is citral (3,7-dimethyl-2,6-octadien-1-al).17,18,19,20,21,22,23,24,25 Lemongrass oil has been found to contain up to 75-85% citral.17,26,21 Lemongrass also contains z-citral, borneol, estragole, methyleugenol, geranyl acetate (3,7-dimethyl-2,6-octadiene-1-ol acetate), geraniol (some species higher in this compound than citral), beta-myrcene (MYR, 7-methyl-3-methylene-1,6 octadiene), limonene, piperitone, citronellal, carene-2, alpha-terpineole, pinene, farnesol, proximadiol, and (+)-cymbodiacetal.11,27,28,29,30,31,32,33,34,35,36,25,37,38,39,40,41,12,42,25 The volatile oil from the roots contains 56.67% longifolene-(V4) and 20.03% selina-6-en-4-ol.25 In particular, a study of Cymbopogon martinii isolated fatty acids, common sterols, and 16-hydroxypentacos-14(z)-enoic acid.43
  • Analgesic effects: Lemongrass has exhibited analgesic effects in mice and rat studies.44,45,46 Intraperitoneal and subcutaneous injections of lemongrass and myrcene inhibited the acetic acid-induced writhing in mice.45,46 Naloxone, an opioid antagonist, blocked the antinociception in the mice, suggesting that the analgesic effect may be a result of the release of endogenous opioids. Oral administration of lemongrass in rats resulted in a dose-dependent analgesia when hyperalgesia is induced by injections of carrageenan or prostaglandin E2, but did not affect the hyperalgesia induced by cyclic AMP, indicating a peripheral site of action.44 The analgesic effect did not cause tolerance on repeated injections in rats. The analgesic component of lemongrass was determined to be myrcene.44,45
  • Anthelmintic effects: Cymbopogon martinii was tested for anthelmintic activity in vitro.47 Geraniol, a constituent of Cymbopogon martinii, showed potent anthelmintic activity and the ED50 of geraniol was found to be 66.7µg/mL.
  • Antibacterial effects: Lemongrass has demonstrated antibacterial properties in laboratory studies.48,49,50,51,52,53,54,55,56,57,58,59,60,61,62,63,64,65,66,67,68,69,70 Some examples of organisms that lemongrass has activity against are Escherichia coli, Salmonella enterica, and Klebsiella pneumoniae.52,53,54,61,65,67 Lemongrass has also demonstrated activity against common respiratory tract pathogens like Haemophilus influenzae, Pseudomonas aeroginosa, Streptococcus aureus, Streptococcus pyogenes, and Staphylococcus aereus.55,57,71,69 One study showed that Cymbopogon citratus completely inhibits the growth of Helicobacter pylori without developing resistance, where clarithromycin developed resistance under the same conditions.60 Lemongrass has also been reported to increase the activity of phenoxythanol against Escherichia coli50,72 and Staphylococcus aureus63.
  • Lemongrass was found to elicit morphological changes, inhibit septum formation, spheroblast formation, production of 'blisters' or mesosomes, development of abnormally shaped cells, and lysis.61 It was also found to permeabilize the cytoplasmic membrane with the leakage of carboxy-fluorescein.59 One study showed that lemongrass might have antibacterial activity by influencing bacterial targets involved in cytoplasmic and cell well metabolism.69
  • Lemongrass was also tested against Escherichia coli wild type (AB 1157) strain submitted to SnCl(2) treatment.72 In the presence of Cymbopogon citratus, the SnCl(2) effect on the survival of Escherichia coli was reduced. Another laboratory study also found that the essential oil from Cymbopogon martinii exhibited broad spectrum inhibiting properties, with strong activity (MIC between 100 and 500mcg/mL) against 10 out of 13 Escherichia coli serotypes: three enterotoxigenic, two enteropathogenic, three enteroinvasive and two shiga-toxin producers. Similarly, C. winterianus significantly inhibited two enterotoxigenic, one enteropathogenic, one enteroinvasive and one shiga-toxin producer serotypes.50
  • Sorrel drink, a combination of roselle calyx aqueous extract, orange or pineapple juice, and lemon grass, had antibacterial effects on Bacillus sp., Pseudomonas aeruginosa, Lactobacillus sp., and Corynebacterium sp., whereas roselle calyx aqueous extract did not show these effects.73
  • Anticancer effects: Cymbopogan citratus Stapf was found to have anti-mutogenic properties in a Salmonella mutation assay.74 Citral, extracted from Cymbopogan citratus, induced death apoptosis, DNA fragmentation, and caspase-3 catalytic activity in several hematopoietic cancer cell lines in anther study.20 In rats, lemongrass extract was administered before inducing DNA adducts and aberrant crypt foci.75 The lemongrass treatment significantly inhibited the DNA adduct formation and aberrant crypt formation in the colon. However, in a study in rats with induced hepatocarcinogenesis, lemongrass extract had only an inhibitory effect on the early phase of hepatocarcinogenesis.76 Limonene, a constituent of lemongrass, is also being tested for its antitumor activity.
  • Antifungal effects: Lemongrass has been shown to have antifungal properties in laboratory studies.77,17,78,79,80,81,82,83,84,85,86,87,88,89,90,91,92,93,94,95,96,97,98,99,100,101,102,103,104,105,106,107
  • Some examples of fungi include various Aspergillus species and Candida albicans. In one study, lemongrass oil was able to treat experimental ringworm in guinea pigs in 7 to 12 days.88 The oil has also been proven effective as a preservative in preventing fungi from growing on foods. There are various proposed antifungal mechanisms of action for lemongrass oil. It has demonstrated an inhibitory effect on hyphal growth and spore formation in Aspergillus niger as well as plasma membrane disruption and mitochondrial structure disorganization.80,85,96 Cymbopogon martinii has also been shown to cause K+ leakage from the yeast cell, as well as changes in the composition in the yeast cell membrane.97
  • Anti-inflammatory effects: In a pharmacological study in mice, anti-inflammatory properties of lemongrass oil was examined with 5microL of lemongrass oil per subject, intraperitoneally.108 Lemongrass oil was shown to suppress the leukocyte recruitment into the peritoneal cavity by inhibiting neutrophil accumulation. In another study, 0.0125 to 0.1% lemongrass oil suppressed neutrophil activation by tumor necrosis factor-alpha (TNF-alpha) in vitro.17
  • Antimalarial effects: Studies in mice have shown that lemongrass is effective against the malaria parasite.109,4,110 One study showed as high as 86.6% response at 500mg/kg of Cymbopogon citratus oil per day in mice where chloroquine 10mg/kg per day in mice was 100% effective.4 Another study in mice proved Cymbopogon giganteus to have anti-malarial activity in chloroquine resistant malaria.109
  • Antimutagenic effects: Constituents of lemongrass have been tested on their antimutagenic effects against cyclophosphamide, mitomycin-C, and nickel chloride.111,112 In one study with mice, citral prevented the nuclear damage induced by cyclophosphamide, mitomycin-C, and nickel chloride. 112
  • Antioxidant effects: Lemongrass oil has been evaluated for its antioxidant properties.113,114,115,116 Citral, a main constituent of lemongrass oil, has been given to rats orally at 60mg/kg for one week.116 Then nickel chloride, a known mutagen, was given intraperitoneally to induce nuclear damage. Citral significantly inhibited the formation of micronuclei induced by nickel when the antioxidant activity of citral was tested in vitro. An EC50 of 19µg/mL was observed in the superoxide scavenging activity for citral, suggesting an anti-clastogenic effect of citral against nickel chloride.
  • Citral was also tested for its induction activity on glutathione S-transferase (GST), which detoxifies polycyclic aromatic hydrocarbons, using rat liver epithelial cell lines.115 Citral dose- and time-dependently induced the activities of GST. Glutathione (GSH) and protein thiol also specifically activated with the isomer geranial. The authors suggest this study implied the antioxidant role of GST induction by citral. Cymbopogon citratus showed free radical scavenging effects in one study and was antigenotoxic against gamma-rays in another study again suggesting free radical scavenging mechanisms.113,114
  • Antiplatelet activity: The essential oil of Cymbopogon citratus has been tested for antiplatelet activity in guinea pig and rat plasma.117
  • Antiviral effects: Lemongrass oil has shown antiviral activity against herpes simplex virus type-1 (HSV-1) in vitro.118 A concentration of 0.1% lemongrass oil completely inhibited viral replication when there was direct interaction with the virions at 4Cº for 24 hours.
  • Cardiac effects: Aqueous extracts of Cymbopogon citratus reduced cardiac rate and did not alter contractile force in isolated hearts of male rats.14 Cymbopogon goeringii volatile oil inhibited the contraction and prolonged the functional refractory period in isolated guinea pig papillary muscles and atrium, suggesting the anti-arrhythmic actions of this species.15 Lemongrass oil also demonstrated vasorelaxation on isolated, perfused, mesenteric artery preparation and appears to be mediated by nitric oxide-independent and non-postanoid mechanisms.13,16 Cymbopogon citratus showed some hypotensive effects that were dose related when given intravenously to rats.5 It also showed some weak diuretic and anti-inflammatory effects when given orally to the rats.
  • CNS effects: An oral infusion of up to 208 times the corresponding human dose of lemongrass and oral citral up to 200mg/kg in rats showed no effect on the central nervous system (CNS) as a depressant, hypnotic, neuroleptic, anti-convulsant, or anxiolytic.6 Intraperitoneal doses also exhibited little or no effect on the CNS of rats. In another study, rats and mice received beta-myrcene, a constituent of lemongrass, 1g/kg orally in corn oil27. This study also showed no effect of lemongrass acting on the central nervous system. There have been other studies conducted on the effects of lemongrass on the CNS.119,120
  • Hypoglycemic effects: There were two studies conducted to determine the hypoglycemic properties of lemongrass (Cymbopogon proximus) in alloxan-induced diabetic rats.9,10 Both studies showed that lemongrass restored glucose levels to normal in four weeks of treatment in rats. In one study, alloxan-induced diabetic rats were given 1.5mL of aqueous suspension of lemongrass per 100g body weight orally each day for four weeks.9 The lemongrass treatment restored glucose, urea, creatinine, bilirubin, total protein and albumin and decreased aspartate aminotransferase (AST), alanine aminotransferase (ALT) and lactate dehydrogenase (LDH) in the alloxan-induced diabetic rats. In the other study, lemongrass was administered orally each day to alloxan-induced diabetic rats and elevated blood glucose returned to normal levels after four weeks.10 Lemongrass treatment also decreased cytochrome P450s, and their associated enzyme activities such as aryl hydrocarbon (benzo(a)pyrene) hydroxylase (AHH), N-nitrosdimethylamine N-demethylase I (NDMA-dI), NADPH-cytochrome C reductase, and detoxified by glutathione S-transferase (GST) and glutathione (GSH) after these enzymes had been increased in alloxan-induced diabetic rats.
  • Insecticide and insect repellent effects: Lemongrass has been shown to have insect repellent effects.3,121,122,123,124,125,126 Various studies have shown lemongrass oil to be anywhere from 50 to 95% effective at repelling insects for two to three hours.3,122,123,126. Lemongrass has also shown insecticidal.2,127,128,129,35,36,130,131,132,133,134 Studies have shown that lemongrass oil is an effective insecticide against various insects like dust mites, termites, and ticks.
  • Other: Lemongrass was tested to determine its affect on 5-HT release for use in headache treatment.1 Cymbopogon ambiguus inhibited adenosine diphosphate (ADP) induced platelet 5-HT release in levels ranging from 56-98% and was not due to the non-specific effects of protein binding tannins. The inhibition was dose dependent with an IC50 value of 0.758mg/mL. The dichloromethane (DCM) extract of C. ambiguus also caused a significant inhibition of platelet activation induced by epinephrine, arachidonic acid, and to a lesser extent by collagen. The authors suggest these results indicate that inhibition may occur within the arachidonic acid pathway and suggest the presence of a cyclo-oxygenase inhibitor.
  • A test was conducted to assess the effect of Cymbopogon citratus on levels of serum aflatoxin-albumin (AF-albumin) adducts following aflatoxin B(1) (AFB(1)) exposure in rats.135 Rats were treated with 5g/kg Cymbopogon citratus extract daily for one week, then the rats were administered AFB(1). No significant difference in biomarker levels was seen with the Cymbopogon citratus extract treatments compared to control rats.
  • An in vitro test found that Cymbopogon densiflorus possessed significant antidrepanocytary activity (anti-sickle cell anemia), supporting the traditional use of this herb by Congolese healers.136
  • An in vitro study found that ethanolic extracts of Cymbopogon citratus had enzyme inhibiting ability (≥50 %) against acetylcholinesterase (AChE), butyrylcholinesterase (BChE), and lipoxygenase enzymes (LO).137
  • Extracts from the lemongrass plant, when reacted with aqueous chloroaurate ions, yields a high percentage of thin, flat, single-crystalline gold nanotriangles used in nanaotechnology.7,8
Pharmacodynamics/Kinetics:
  • The urinary metabolites of citral, the main constituent of lemongrass oil (up to 80% of lemongrass oil)23, was studied in male rats138. Urine was collected for 24 hours after a single oral citral dose of 500mg/kg. The elimination in the urine was found to be rapid, with approximately 50% of the dose excreted within 24 hours. Citral was rapidly metabolized and excreted as metabolites. The metabolites were a biliary glucuronide and several acids. The metabolites that were isolated were 3-hydroxy-3,7-dimethyl-6-octenedioic acid; 3,8-dihydroxy-3,7-dimethyl-6-octenoic acid; 3,9-dihydroxy-3,7-dimethyl-6-octenoic acid; E- and Z-3,7-dimethyl-2,6-octadienedioic acid; 3,7-dimethyl-6-octenedioic acid; and E-3,7-dimethyl-2,6-octadienoic acid. The urinary metabolites of citral appeared to arise from metabolic pathways other than a nucleophilic addition to the double bond.
References
  1. Rogers, K. L., Grice, I. D., and Griffiths, L. R. Modulation of in vitro platelet 5-HT release by species of Erythrina and Cymbopogon. Life Sci 8-31-2001;69(15):1817-1829. 11665843
  2. Bassole, I. H., Guelbeogo, W. M., Nebie, R., Costantini, C., Sagnon, N., Kabore, Z. I., and Traore, S. A. Ovicidal and larvicidal activity against Aedes aegypti and Anopheles gambiae complex mosquitoes of essential oils extracted from three spontaneous plants of Burkina Faso. Parassitologia 2003;45(1):23-26. 15270540
  3. Ansari, M. A. and Razdan, R. K. Relative efficacy of various oils in repelling mosquitoes. Indian J Malariol 1995;32(3):104-111. 8936292
  4. Tchoumbougnang, F., Zollo, P. H., Dagne, E., and Mekonnen, Y. In vivo antimalarial activity of essential oils from Cymbopogon citratus and Ocimum gratissimum on mice infected with Plasmodium berghei. Planta Med 2005;71(1):20-23. 15678368
  5. Carbajal, D., Casaco, A., Arruzazabala, L., Gonzalez, R., and Tolon, Z. Pharmacological study of Cymbopogon citratus leaves. J Ethnopharmacol 1989;25(1):103-107. 2716341
  6. Carlini, E. A., Contar, J. de DP, Silva-Filho, A. R., Silveira-Filho, N. G., Frochtengarten, M. L., and Bueno, O. F. Pharmacology of lemongrass (Cymbopogon citratus Stapf). I. Effects of teas prepared from the leaves on laboratory animals. J Ethnopharmacol 1986;17(1):37-64. 3762195
  7. Rai, A., Singh, A., Ahmad, A., and Sastry, M. Role of halide ions and temperature on the morphology of biologically synthesized gold nanotriangles. Langmuir 1-17-2006;22(2):736-741. 16401125
  8. Shankar, S. S., Rai, A., Ankamwar, B., Singh, A., Ahmad, A., and Sastry, M. Biological synthesis of triangular gold nanoprisms. Nat Mater 2004;3(7):482-488. 15208703
  9. Mansour, H. A., Newairy, A. S., Yousef, M. I., and Sheweita, S. A. Biochemical study on the effects of some Egyptian herbs in alloxan-induced diabetic rats. Toxicology 1-25-2002;170(3):221-228. 11788159
  10. Sheweita, S. A., Newairy, A. A., Mansour, H. A., and Yousef, M. I. Effect of some hypoglycemic herbs on the activity of phase I and II drug-metabolizing enzymes in alloxan-induced diabetic rats. Toxicology 5-24-2002;174(2):131-139. 11985890
  11. NTP Carcinogenesis Studies of Food Grade Geranyl Acetate (71% Geranyl Acetate, 29% Citronellyl Acetate) (CAS No. 105-87-3) in F344/N Rats and B6C3F1 Mice (Gavage Study). Natl Toxicol Program Tech Rep SerM 1987;252:1-162. 12748693
  12. Zamith, H. P., Vidal, M. N., Speit, G., and Paumgartten, F. J. Absence of genotoxic activity of beta-myrcene in the in vivo cytogenetic bone marrow assay. Braz J Med Biol Res 1993;26(1):93-98. 8220273
  13. Abeywardena, M., Runnie, I., Nizar, M., Suhaila, M., and Head, R. Polyphenol-enriched extract of oil palm fronds (Elaeis guineensis) promotes vascular relaxation via endothelium-dependent mechanisms. Asia Pac J Clin Nutr 2002;11 Suppl 7:S467-S472. 12492636
  14. Gazola, R., Machado, D., Ruggiero, C., Singi, G., and Macedo, Alexandre M. Lippia alba, Melissa officinalis and Cymbopogon citratus: effects of the aqueous extracts on the isolated hearts of rats. Pharmacol Res 2004;50(5):477-480. 15458767
  15. Liu, M. and Feng, G. H. [Effects of Cymbopogon goeringii (Steud.) A. Camus volatile oil on physiologic properties of the isolated guinea pig myocardium]. Zhongguo Zhong Yao Za Zhi 1989;14(10):620-3, 640. 2597328
  16. Runnie, I., Salleh, M. N., Mohamed, S., Head, R. J., and Abeywardena, M. Y. Vasorelaxation induced by common edible tropical plant extracts in isolated rat aorta and mesenteric vascular bed. J Ethnopharmacol 2004;92(2-3):311-316. 15138017
  17. Abe, S., Sato, Y., Inoue, S., Ishibashi, H., Maruyama, N., Takizawa, T., Oshima, H., and Yamaguchi, H. [Anti-Candida albicans activity of essential oils including Lemongrass (Cymbopogon citratus) oil and its component, citral]. Nippon Ishinkin Gakkai Zasshi 2003;44(4):285-291. 14615795
  18. Balbaa, S. I. and Johnson, C. H. The microscopic structure of lemongrass leaves. J Am Pharm Assoc Am Pharm Assoc (Baltim) 1955;44(2):89-98. 14353722
  19. Banthorpe, D. V., Duprey, R. J., Hassan, M., Janes, J. F., and Modawi, B. M. Chemistry of the Sudanese flora. I. Essential oils of some Cymbopogon species. Planta Med 1976;29(1):10-25. 1257323
  20. Dudai, N., Weinstein, Y., Krup, M., Rabinski, T., and Ofir, R. Citral is a new inducer of caspase-3 in tumor cell lines. Planta Med 2005;71(5):484-488. 15931590
  21. Rauber, Cda S., Guterres, S. S., and Schapoval, E. E. LC determination of citral in Cymbopogon citratus volatile oil. J Pharm Biomed Anal. 3-9-2005;37(3):597-601. 15740922
  22. Saleem, M., Afza, N., Anwar, M. A., Hai, S. M., and Ali, M. S. A comparative study of essential oils of Cymbopogon citratus and some members of the genus Citrus. Nat Prod Res 2003;17(5):369-373. 14526919
  23. Saleem, M., Afza, N., Anwar, M. A., Hai, S. M., Ali, M. S., Shujaat, S., and Atta, Ur rahman. Chemistry and biological significance of essential oils of Cymbopogon citratus from Pakistan. Nat Prod Res 2003;17(3):159-163. 12737398
  24. Wilson, N. D., Ivanova, M. S., Watt, R. A., and Moffat, A. C. The quantification of citral in lemongrass and lemon oils by near-infrared spectroscopy. J Pharm Pharmacol 2002;54(9):1257-1263. 12356280
  25. Li, H., Huang, J., Zhang, X., Chen, Y., Yang, J., and Hei, L. [Allelopathic effects of Cymbopogon citratu volatile and its chemical components]. Ying Yong Sheng Tai Xue Bao 2005;16(4):763-767. 16011184
  26. Nhu-Trang, T. T., Casabianca, H., and Grenier-Loustalot, M. F. Authenticity control of essential oils containing citronellal and citral by chiral and stable-isotope gas-chromatographic analysis. Anal Bioanal Chem 2006;386(7-8):2141-2152. 17089103
  27. da Silva, V. A., de Freitas, J. C., Mattos, A. P., Paiva-Gouvea, W., Presgrave, O. A., Fingola, F. F., Menezes, M. A., and Paumgartten, F. J. Neurobehavioral study of the effect of beta-myrcene on rodents. Braz J Med Biol Res 1991;24(8):827-831. 1797273
  28. D'Souza, A. M., Paknikar, S. K., Dev, V., Beauchamp, P. S., and Kamat, S. P. Biogenetic-type synthesis of (+)-cymbodiacetal, a constituent of Cymbopogon martinii. J Nat Prod. 2004;67(4):700-702. 15104509
  29. Dubey, V. S., Bhalla, R., and Luthra, R. An esterase is involved in geraniol production during palmarosa inflorescence development. Phytochemistry 2003;63(3):257-264. 12737976
  30. Dubey, V. S., Bhalla, R., and Luthra, R. Sucrose mobilization in relation to essential oil biogenesis during palmarosa (Cymbopogon martinii Roxb. Wats. var. motia) inflorescence development. J Biosci. 2003;28(4):479-487. 12799494
  31. Dubey, V. S. and Luthra, R. Biotransformation of geranyl acetate to geraniol during palmarosa (Cymbopogon martinii, Roxb. wats. var. motia) inflorescence development. Phytochemistry 2001;57(5):675-680. 11397433
  32. Dudai, N., Weinberg, Z. G., Larkov, O., Ravid, U., Ashbell, G., and Putievsky, E. Changes in essential oil during enzyme-assisted ensiling of lemongrass (Cymbopogon citratus Stapf.) and lemon eucalyptus (Eucalyptus citriodora Hook). J Agric Food Chem 2001;49(5):2262-2266. 11368586
  33. Fujita, S. I. and Fujita, Y. [Miscellaneous contributions to the essential oils of the plants from various territories. XXX. On the components of the essential oils of Cymbopogon goeringii A. Camus]. Yakugaku Zasshi 1972;92(10):1285-1288. 4674731
  34. Hegnauer, R. [Microscopical study of the leaves of Cymbopogon flexuosus Stapf (lemon grass).]. Pharm Weekbl 6-18-1955;90(13):445-447. 14394824
  35. Ketoh, G. K., Glitho, A. I., and Huignard, J. Susceptibility of the bruchid Callosobruchus maculatus (Coleoptera: Bruchidae) and its parasitoid Dinarmus basalis (Hymenoptera: Pteromalidae) to three essential oils. J Econ Entomol 2002;95(1):174-182. 11942754
  36. Ketoh, G. K., Koumaglo, H. K., Glitho, I. A., and Huignard, J. Comparative effects of Cymbopogon schoenanthus essential oil and piperitone on Callosobruchus maculatus development. Fitoterapia 7-16-2006;16938411
  37. Marongiu, B., Piras, A., Porcedda, S., and Tuveri, E. Comparative analysis of the oil and supercritical CO(2) extract of Cymbopogon citratus Stapf. Nat Prod Res 5-10-2006;20(5):455-459. 16644543
  38. Matsuda, B. M., Surgeoner, G. A., Heal, J. D., Tucker, A. O., and Maciarello, M. J. Essential oil analysis and field evaluation of the citrosa plant "Pelargonium citrosum" as a repellent against populations of Aedes mosquitoes. J Am Mosq Control Assoc 1996;12(1):69-74. 8723261
  39. Naves, Y. R. and Grampoloff, A. V. [p-Menthadienols in the essential oils of Cymbopogon martini Stapf var. sofia (essence of gingergrass) and of Cymbopogon densiflorus Stapf.]. C.R. Hebd Seances Acad Sci 7-15-1959;249(2):306-308. 13671817
  40. Radwan, A. S. An analytical method for proximadiol, the active principle of Cymbopogon proximus. Planta Med 1975;27(1):93-97. 1161881
  41. Schaneberg, B. T. and Khan, I. A. Comparison of extraction methods for marker compounds in the essential oil of lemon grass by GC. J Agric Food Chem 3-13-2002;50(6):1345-1349. 11879000
  42. Iten, F. and Saller, R. [Fennel tea: risk assessment of the phytogenic monosubstance estragole in comparison to the natural multicomponent mixture]. Forsch Komplementarmed Klass Naturheilkd 2004;11(2):104-108. 15138375
  43. Siddiqi, S. A. and Misra, L. Fatty acids and sterols from Cymbopogon martinii var. motia roots. Z Naturforsch [C] 2000;55(9-10):843-845. 11098842
  44. Lorenzetti, B. B., Souza, G. E., Sarti, S. J., Santos, Filho D., and Ferreira, S. H. Myrcene mimics the peripheral analgesic activity of lemongrass tea. J Ethnopharmacol 1991;34(1):43-48. 1753786
  45. Rao, V. S., Menezes, A. M., and Viana, G. S. Effect of myrcene on nociception in mice. J Pharm Pharmacol 1990;42(12):877-878. 1753786
  46. Viana, G. S., Vale, T. G., Pinho, R. S., and Matos, F. J. Antinociceptive effect of the essential oil from Cymbopogon citratus in mice. J Ethnopharmacol 2000;70(3):323-327. 10837994
  47. Kumaran, A. M., D'Souza, P., Agarwal, A., Bokkolla, R. M., and Balasubramaniam, M. Geraniol, the putative anthelmintic principle of Cymbopogon martinii. Phytother Res 2003;17(8):957. 13680833
  48. Betoni, J. E., Mantovani, R. P., Barbosa, L. N., Di Stasi, L. C., and Fernandes, Junior A. Synergism between plant extract and antimicrobial drugs used on Staphylococcus aureus diseases. Mem Inst Oswaldo Cruz 2006;101(4):387-390. 16951808
  49. Cimanga, K., Kambu, K., Tona, L., Apers, S., De Bruyne, T., Hermans, N., Totte, J., Pieters, L., and Vlietinck, A. J. Correlation between chemical composition and antibacterial activity of essential oils of some aromatic medicinal plants growing in the Democratic Republic of Congo. J Ethnopharmacol 2002;79(2):213-220. 11801384
  50. Duarte, M. C., Leme, E. E., Delarmelina, C., Soares, A. A., Figueira, G. M., and Sartoratto, A. Activity of essential oils from Brazilian medicinal plants on Escherichia coli. J Ethnopharmacol 12-3-2006;17210236
  51. Dorman, H. J. and Deans, S. G. Antimicrobial agents from plants: antibacterial activity of plant volatile oils. J Appl Microbiol 2000;88(2):308-316. 10736000
  52. El Kamali, H. H., Hamza, M. A., and El Amir, M. Y. Antibacterial activity of the essential oil from Cymbopogon nervatus inflorescence. Fitoterapia 2005;76(5):446-449. 15896926
  53. Friedman, M., Henika, P. R., Levin, C. E., and Mandrell, R. E. Antibacterial activities of plant essential oils and their components against Escherichia coli O157:H7 and Salmonella enterica in apple juice. J Agric Food Chem 9-22-2004;52(19):6042-6048. 15366861
  54. Friedman, M., Henika, P. R., and Mandrell, R. E. Bactericidal activities of plant essential oils and some of their isolated constituents against Campylobacter jejuni, Escherichia coli, Listeria monocytogenes, and Salmonella enterica. J Food Prot 2002;65(10):1545-1560. 12380738
  55. Inouye, S., Takizawa, T., and Yamaguchi, H. Antibacterial activity of essential oils and their major constituents against respiratory tract pathogens by gaseous contact. J Antimicrob Chemother 2001;47(5):565-573. 11328766
  56. Inouye, S., Uchida, K., and Yamaguchi, H. In-vitro and in-vivo anti-Trichophyton activity of essential oils by vapour contact. Mycoses 2001;44(3-4):99-107. 11413931
  57. Inouye, S., Yamaguchi, H., and Takizawa, T. Screening of the antibacterial effects of a variety of essential oils on respiratory tract pathogens, using a modified dilution assay method. J Infect Chemother 2001;7(4):251-254. 11810593
  58. Mejlholm, O. and Dalgaard, P. Antimicrobial effect of essential oils on the seafood spoilage micro-organism Photobacterium phosphoreum in liquid media and fish products. Lett Appl Microbiol 2002;34(1):27-31. 11849488
  59. Nguefack, J., Budde, B. B., and Jakobsen, M. Five essential oils from aromatic plants of Cameroon: their antibacterial activity and ability to permeabilize the cytoplasmic membrane of Listeria innocua examined by flow cytometry. Lett Appl Microbiol 2004;39(5):395-400. 15482428
  60. Ohno, T., Kita, M., Yamaoka, Y., Imamura, S., Yamamoto, T., Mitsufuji, S., Kodama, T., Kashima, K., and Imanishi, J. Antimicrobial activity of essential oils against Helicobacter pylori. Helicobacter 2003;8(3):207-215. 12752733
  61. Ogunlana, E. O., Hoglund, S., Onawunmi, G., and Skold, O. Effects of lemongrass oil on the morphological characteristics and peptidoglycan synthesis of Escherichia coli cells. Microbios 1987;50(202):43-59. 3299005
  62. Onawunmi, G. O., Yisak, W. A., and Ogunlana, E. O. Antibacterial constituents in the essential oil of Cymbopogon citratus (DC.) Stapf. J Ethnopharmacol 1984;12(3):279-286. 6442749
  63. Onawunmi, G. O. In vitro studies on the antibacterial activity of phenoxyethanol in combination with lemon grass oil. Pharmazie 1988;43(1):42-44. 3287398
  64. Pattnaik, S., Subramanyam, V. R., Kole, C. R., and Sahoo, S. Antibacterial activity of essential oils from Cymbopogon: inter- and intra-specific differences. Microbios 1995;84(341):239-245. 8643027
  65. Pattnaik, S., Subramanyam, V. R., and Rath, C. C. Effect of essential oils on the viability and morphology of Escherichia coli (SP-11). Microbios 1995;84(340):195-199. 8820244
  66. Raybaudi-Massilia, R. M., Mosqueda-Melgar, J., and Martin-Belloso, O. Antimicrobial activity of essential oils on Salmonella enteritidis, Escherichia coli, and Listeria innocua in fruit juices. J Food Prot 2006;69(7):1579-1586. 16865889
  67. Rojas-Grau, M. A., Avena-Bustillos, R. J., Friedman, M., Henika, P. R., Martin-Belloso, O., and McHugh, T. H. Mechanical, barrier, and antimicrobial properties of apple puree edible films containing plant essential oils. J Agric Food Chem 11-29-2006;54(24):9262-9267. 17117819
  68. Takaisi-Kikuni, N. B., Tshilanda, D., and Babady, B. Antibacterial activity of the essential oil of Cymbopogon densiflorus. Fitoterapia 2000;71(1):69-71. 11449474
  69. Takaisi-Kikuni, N. B., Kruger, D., Gnann, W., and Wecke, J. Microcalorimetric and electron microscopic investigation on the effects of essential oil from Cymbopogon densiflorus on Staphylococcus aureus. Microbios 1996;88(354):55-62. 9121380
  70. Wannissorn, B., Jarikasem, S., Siriwangchai, T., and Thubthimthed, S. Antibacterial properties of essential oils from Thai medicinal plants. Fitoterapia 2005;76(2):233-236. 15752638
  71. Pattnaik, S., Rath, C., and Subramanyam, V. R. Characterization of resistance to essential oils in a strain of Pseudomonas aeruginosa (VR-6). Microbios 1995;81(326):29-31. 7731390
  72. Melo, S. F., Soares, S. F., da Costa, R. F., da Silva, C. R., de Oliveira, M. B., Bezerra, R. J., Caldeira-de-Araujo, A., and Bernardo-Filho, M. Effect of the Cymbopogon citratus, Maytenus ilicifolia and Baccharis genistelloides extracts against the stannous chloride oxidative damage in Escherichia coli. Mutat Res 9-20-2001;496(1-2):33-38. 11551478
  73. Oboh, G. and Elusiyan, C. A. Nutrient composition and antimicrobial activity of sorrel drinks (soborodo). J Med Food 2004;7(3):340-342. 15383229
  74. Vinitketkumnuen, U., Puatanachokchai, R., Kongtawelert, P., Lertprasertsuke, N., and Matsushima, T. Antimutagenicity of lemon grass (Cymbopogon citratus Stapf) to various known mutagens in salmonella mutation assay. Mutat Res 1994;341(1):71-75. 7523944
  75. Suaeyun, R., Kinouchi, T., Arimochi, H., Vinitketkumnuen, U., and Ohnishi, Y. Inhibitory effects of lemon grass (Cymbopogon citratus Stapf) on formation of azoxymethane-induced DNA adducts and aberrant crypt foci in the rat colon. Carcinogenesis 1997;18(5):949-955. 9163680
  76. Puatanachokchai, R., Kishida, H., Denda, A., Murata, N., Konishi, Y., Vinitketkumnuen, U., and Nakae, D. Inhibitory effects of lemon grass (Cymbopogon citratus, Stapf) extract on the early phase of hepatocarcinogenesis after initiation with diethylnitrosamine in male Fischer 344 rats. Cancer Lett 9-8-2002;183(1):9-15. 12049809
  77. Essential oils of peppermint, orange or lemongrass kill most strains of fungal and bacterial infections. Posit Health News 1998;(No 17):26-27. 11366554
  78. Anthony, S., Abeywickrama, K., Dayananda, R., Wijeratnam, S. W., and Arambewela, L. Fungal pathogens associated with banana fruit in Sri Lanka, and their treatment with essential oils. Mycopathologia 2004;157(1):91-97. 15008351
  79. Bankole, S. A., Joda, A. O., and Ashidi, J. S. The use of powder and essential oil of Cymbopogon citratus against mould deterioration and aflatoxin contamination of "egusi" melon seeds. J Basic Microbiol 2005;45(1):20-30. 15678559
  80. de Billerbeck, V. G., Roques, C. G., Bessiere, J. M., Fonvieille, J. L., and Dargent, R. Effects of Cymbopogon nardus (L.) W. Watson essential oil on the growth and morphogenesis of Aspergillus niger. Can J Microbiol 2001;47(1):9-17. 15049444
  81. Duarte, M. C., Figueira, G. M., Sartoratto, A., Rehder, V. L., and Delarmelina, C. Anti-Candida activity of Brazilian medicinal plants. J Ethnopharmacol 2-28-2005;97(2):305-311. 15707770
  82. Fandohan, P., Gbenou, J. D., Gnonlonfin, B., Hell, K., Marasas, W. F., and Wingfield, M. J. Effect of essential oils on the growth of Fusarium verticillioides and fumonisin contamination in corn. J Agric Food Chem 11-3-2004;52(22):6824-6829. 15506822
  83. Guynot, M. E., Ramos, A. J., Seto, L., Purroy, P., Sanchis, V., and Marin, S. Antifungal activity of volatile compounds generated by essential oils against fungi commonly causing deterioration of bakery products. J Appl Microbiol 2003;94(5):893-899. 12694455
  84. Hammer, K. A., Carson, C. F., and Riley, T. V. Antimicrobial activity of essential oils and other plant extracts. J Appl Microbiol 1999;86(6):985-990. 10438227
  85. Helal, G. A., Sarhan, M. M., Abu Shahla, A. N., and Abou El-Khair, E. K. Effects of Cymbopogon citratus L. essential oil on the growth, lipid content and morphogenesis of Aspergillus niger ML2-strain. J Basic Microbiol 2006;46(6):456-469. 17139611
  86. Inouye, S., Tsuruoka, T., Watanabe, M., Takeo, K., Akao, M., Nishiyama, Y., and Yamaguchi, H. Inhibitory effect of essential oils on apical growth of Aspergillus fumigatus by vapour contact. Mycoses 2000;43(1-2):17-23. 10838841
  87. Inouye, S., Watanabe, M., Nishiyama, Y., Takeo, K., Akao, M., and Yamaguchi, H. Antisporulating and respiration-inhibitory effects of essential oils on filamentous fungi. Mycoses 1998;41(9-10):403-410. 9916464
  88. Kishore, N., Mishra, A. K., and Chansouria, J. P. Fungitoxicity of essential oils against dermatophytes. Mycoses 1993;36(5-6):211-215. 8264720
  89. Lima, E. O., Gompertz, O. F., Giesbrecht, A. M., and Paulo, M. Q. In vitro antifungal activity of essential oils obtained from officinal plants against dermatophytes. Mycoses 1993;36(9-10):333-336. 8015567
  90. Mishra, A. K. and Dubey, N. K. Evaluation of some essential oils for their toxicity against fungi causing deterioration of stored food commodities. Appl Environ Microbiol 1994;60(4):1101-1105. 8017906
  91. Nguefack, J., Leth, V., Amvam Zollo, P. H., and Mathur, S. B. Evaluation of five essential oils from aromatic plants of Cameroon for controlling food spoilage and mycotoxin producing fungi. Int J Food Microbiol 8-1-2004;94(3):329-334. 15246244
  92. Palhano, F. L., Vilches, T. T., Santos, R. B., Orlando, M. T., Ventura, J. A., and Fernandes, P. M. Inactivation of Colletotrichum gloeosporioides spores by high hydrostatic pressure combined with citral or lemongrass essential oil. Int J Food Microbiol 8-15-2004;95(1):61-66. 15240075
  93. Paranagama, P. A., Abeysekera, K. H., Abeywickrama, K., and Nugaliyadde, L. Fungicidal and anti-aflatoxigenic effects of the essential oil of Cymbopogon citratus (DC.) Stapf. (lemongrass) against Aspergillus flavus Link. isolated from stored rice. Lett Appl Microbiol 2003;37(1):86-90. 12803563
  94. Pattnaik, S., Subramanyam, V. R., and Kole, C. R. Antifungal activity of essential oils from Cymbopogon: inter- and intraspecific differences. Cytobios 1999;97(386):153-159. 10610298
  95. Pattnaik, S., Subramanyam, V. R., and Kole, C. Antibacterial and antifungal activity of ten essential oils in vitro. Microbios 1996;86(349):237-246. 8893526
  96. Pawar, V. C. and Thaker, V. S. In vitro efficacy of 75 essential oils against Aspergillus niger. Mycoses 2006;49(4):316-323. 16784447
  97. Prashar, A., Hili, P., Veness, R. G., and Evans, C. S. Antimicrobial action of palmarosa oil (Cymbopogon martinii) on Saccharomyces cerevisiae. Phytochemistry 2003;63(5):569-575. 12809717
  98. Qureshi, S., Rai, M. K., and Agrawal, S. C. In vitro evaluation of inhibitory nature of extracts of 18-plant species of Chhindwara against 3-keratinophilic fungi. Hindustan Antibiot Bull 1997;39(1-4):56-60. 10386016
  99. Ramos, Ruiz A., De la Torre, R. A., Alonso, N., Villaescusa, A., Betancourt, J., and Vizoso, A. Screening of medicinal plants for induction of somatic segregation activity in Aspergillus nidulans. J Ethnopharmacol 7-5-1996;52(3):123-127. 8771452
  100. Rolz, C., de Leon, R., de Arriola, M. C., and de Cabrera, S. Biodelignification of Lemon Grass and Citronella Bagasse by White-Rot Fungi. Appl Environ Microbiol  1986;52(4):607-611. 16347155
  101. Sarbhoy, A. K., Varshney, J. L., Maheshwari, M. L., and Saxena, D. B. Efficacy of some essential oils and their constituents on few ubiquitous molds. Zentralbl Bakteriol Naturwiss 1978;133(7-8):723-725. 749414
  102. Suhr, K. I. and Nielsen, P. V. Antifungal activity of essential oils evaluated by two different application techniques against rye bread spoilage fungi. J Appl Microbiol 2003;94(4):665-674. 12631202
  103. Tampieri, M. P., Galuppi, R., Macchioni, F., Carelle, M. S., Falcioni, L., Cioni, P. L., and Morelli, I. The inhibition of Candida albicans by selected essential oils and their major components. Mycopathologia 2005;159(3):339-345. 15883716
  104. Tanu, Prakash, A., and Adholeya, A. Effect of different organic manures/composts on the herbage and essential oil yield of Cymbopogon winterianus and their influence on the native AM population in a marginal alfisol. Bioresour Technol 2004;92(3):311-319. 14766166
  105. Velluti, A., Sanchis, V., Ramos, A. J., Egido, J., and Marin, S. Inhibitory effect of cinnamon, clove, lemongrass, oregano and palmarose essential oils on growth and fumonisin B1 production by Fusarium proliferatum in maize grain. Int J Food Microbiol 12-31-2003;89(2-3):145-154. 14623380
  106. Velluti, A., Sanchis, V., Ramos, A. J., Turon, C., and Marin, S. Impact of essential oils on growth rate, zearalenone and deoxynivalenol production by Fusarium graminearum under different temperature and water activity conditions in maize grain. J Appl Microbiol 2004;96(4):716-724. 15012810
  107. Wannissorn, B., Jarikasem, S., and Soontorntanasart, T. Antifungal activity of lemon grass oil and lemon grass oil cream. Phytother Res 1996;10:551.
  108. Abe, S., Maruyama, N., Hayama, K., Inouye, S., Oshima, H., and Yamaguchi, H. Suppression of neutrophil recruitment in mice by geranium essential oil. Mediators Inflamm 2004;13(1):21-24. 15203560
  109. Kimbi, H. K. and Fagbenro-Beyioku, A. F. Efficacy of Cymbopogon giganteus and Enantia chrantha against chloroquine resistant Plasmodium yoelii nigeriensis. East Afr Med J 1996;73(10):636-637. 8997841
  110. Williams, G. O. and Omoh, L. E. Mitotic effects of the aqueous leaf extract of Cymbopogon citratus in Allium cepa root tips. Cytobios 1996;87(350):161-168. 9172394
  111. Kauderer, B., Zamith, H., Paumgartten, F. J., and Speit, G. Evaluation of the mutagenicity of beta-myrcene in mammalian cells in vitro. Environ Mol Mutagen 1991;18(1):28-34. 1864266
  112. Rabbani, S. I., Devi, K., and Zahra, N. Anti-clastogenic effects of citral. Iranian Journal of Pharmacology and Therapuetics 2005;4:28-31.
  113. Cheel, J., Theoduloz, C., Rodriguez, J., and Schmeda-Hirschmann, G. Free radical scavengers and antioxidants from Lemongrass (Cymbopogon citratus (DC.) Stapf.). J Agric Food Chem 4-6-2005;53(7):2511-2517. 15796587
  114. Fuentes, J. L., Alonso, A., Cuetara, E., Vernhe, M., Alvarez, N., Sanchez-Lamar, A., and Llagostera, M. Usefulness of the SOS Chromotest in the study of medicinal plants as radioprotectors. Int J Radiat Biol 2006;82(5):323-329. 16782649
  115. Nakamura, Y., Miyamoto, M., Murakami, A., Ohigashi, H., Osawa, T., and Uchida, K. A phase II detoxification enzyme inducer from lemongrass: identification of citral and involvement of electrophilic reaction in the enzyme induction. Biochem Biophys Res Commun 3-14-2003;302(3):593-600. 12615076
  116. Rabbani, S. I., Devi, K., Khanam, S., and Zahra, N. Citral, a component of lemongrass oil inhibits the clastogenic effect of nickel chloride in mouse micronucleus test system. Pak J Pharm Sci 2006;19(2):108-113. 16751120
  117. Tognolini, M., Barocelli, E., Ballabeni, V., Bruni, R., Bianchi, A., Chiavarini, M., and Impicciatore, M. Comparative screening of plant essential oils: phenylpropanoid moiety as basic core for antiplatelet activity. Life Sci 2-23-2006;78(13):1419-1432. 16274702
  118. Minami, M., Kita, M., Nakaya, T., Yamamoto, T., Kuriyama, H., and Imanishi, J. The inhibitory effect of essential oils on herpes simplex virus type-1 replication in vitro. Microbiol Immunol 2003;47(9):681-684. 14584615
  119. Kokate, C. K., Rao, R. E., and Varma, K. C. Pharmacological investigations of essential oil of Cymbopogon nardus (L.) Rendle: studies on central nervous system. Indian J Exp Biol 1971;9(4):515-516. 5147185
  120. Seth, G., Kokate, C. K., and Varma, K. C. Effect of essential oil of cymbopogon citratus stapf. on central nervous system. Indian J Exp Biol 1976;14(3):370-371. 992793
  121. Gilbert, B., Teixeira, D. F., Carvalho, E. S., De Paula, A. E., Pereira, J. F., Ferreira, J. L., Almeida, M. B., Machado, Rda S., and Cascon, V. Activities of the Pharmaceutical Technology Institute of the Oswaldo Cruz Foundation with medicinal, insecticidal and insect repellent plants. An Acad Bras Cienc 1999;71(2):265-271. 10412493
  122. Govere, J., Durrheim, D. N., Du, Toit N., Hunt, R. H., and Coetzee, M. Local plants as repellents against Anopheles arabiensis, in Mpumalanga Province, South Africa. Cent Afr J Med 2000;46(8):213-216. 11317593
  123. Oyedele, A. O., Gbolade, A. A., Sosan, M. B., Adewoyin, F. B., Soyelu, O. L., and Orafidiya, O. O. Formulation of an effective mosquito-repellent topical product from lemongrass oil. Phytomedicine 2002;9(3):259-262. 12046869
  124. Scott, I. M., Jensen, H., Scott, J. G., Isman, M. B., Arnason, J. T., and Philogene, B. J. Botanical insecticides for controlling agricultural pests: piperamides and the Colorado Potato Beetle Leptinotarsa decemlineata say (Coleoptera: Chrysomelidae). Arch Insect Biochem Physiol 2003;54(4):212-225. 14635182
  125. Tawatsin, A., Wratten, S. D., Scott, R. R., Thavara, U., and Techadamrongsin, Y. Repellency of volatile oils from plants against three mosquito vectors. J Vector Ecol 2001;26(1):76-82. 11469188
  126. Trongtokit, Y., Rongsriyam, Y., Komalamisra, N., and Apiwathnasorn, C. Comparative repellency of 38 essential oils against mosquito bites. Phytother Res 2005;19(4):303-309. 16041723
  127. Cavalcanti, E. S., Morais, S. M., Lima, M. A., and Santana, E. W. Larvicidal activity of essential oils from Brazilian plants against Aedes aegypti L. Mem Inst Oswaldo Cruz 2004;99(5):541-544. 15543421
  128. de Mendonca, F. A., da Silva, K. F., dos Santos, K. K., Ribeiro Junior, K. A., and Sant'Ana, A. E. Activities of some Brazilian plants against larvae of the mosquito Aedes aegypti. Fitoterapia 2005;76(7-8):629-636. 16253435
  129. Heimerdinger, A., Olivo, C. J., Molento, M. B., Agnolin, C. A., Ziech, M. F., Scaravelli, L. F., Skonieski, F. R., Both, J. F., and Charao, P. S. [Alcoholic extract of lemongrass (Cymbopogon citratus) on the control of Boophilus microplus in cattle]. Rev Bras Parasitol Vet 2006;15(1):37-39. 16647001
  130. Morsy, T. A., Mazyad, S. A., and el Sharkawy, I. M. The larvicidal activity of solvent extracts of three medicinal plants against third instar larvae of Chrysomyia albiceps. J Egypt Soc Parasitol 1998;28(3):699-709. 9914694
  131. Raja, N., Albert, S., Ignacimuthu, S., and Dorn, S. Effect of plant volatile oils in protecting stored cowpea Vigna unguiculata (L.) Walpers against Callosobruchus maculatus (F.) (Coleoptera: Bruchidae) infestation. J Stored Prod Res 2001;37(2):127-132. 11124376
  132. Rim, I. S. and Jee, C. H. Acaricidal effects of herb essential oils against Dermatophagoides farinae and D. pteronyssinus (Acari: Pyroglyphidae) and qualitative analysis of a herb Mentha pulegium(pennyroyal). Korean J Parasitol 2006;44(2):133-138. 16809961
  133. Tiwari, B. K., Bajpai, V. N., and Agarwal, P. N. Evaluation of insecticidal, fumigant and repellent properties of lemongrass oil. Indian J Exp Biol 1966;4(2):128-129. 4380199
  134. Zhu, B. C., Henderson, G., Chen, F., Fei, H., and Laine, R. A. Evaluation of vetiver oil and seven insect-active essential oils against the Formosan subterranean termite. J Chem Ecol 2001;27(8):1617-1625. 11521400
  135. Vinitketkumnuen, U., Chewonarin, T., Dhumtanom, P., Lertprasertsuk, N., and Wild, C. P. Aflatoxin-albumin adduct formation after single and multiple doses of aflatoxin B(1) in rats treated with Thai medicinal plants. Mutat Res 7-16-1999;428(1-2):345-351. 10518006
  136. Mpiana, P. T., Tshibangu, D. S., Shetonde, O. M., and Ngbolua, K. N. In vitro antidrepanocytary actvity (anti-sickle cell anemia) of some congolese plants. Phytomedicine 11-16-2006;17113273
  137. Khattak, S., Saeed, Ur Rehman, Shah, H. U., Khan, T., and Ahmad, M. In vitro enzyme inhibition activities of crude ethanolic extracts derived from medicinal plants of Pakistan. Nat Prod Res 2005;19(6):567-571. 16010821
  138. Diliberto, J. J., Srinivas, P., Overstreet, D., Usha, G., Burka, L. T., and Birnbaum, L. S. Metabolism of citral, an alpha,beta-unsaturated aldehyde, in male F344 rats. Drug Metab Dispos 1990;18(6):866-875. 1981530
Licensed by Natural Standard Copyright © 2010 by Natural Standard Corporation. All Rights Reserved.
back to Plant Profiler
back to top