Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Nutrition Research > Learning Center > Plant Profiler > California poppy (Eschscholzia californica)
Plant Profiler
California poppy (Eschscholzia californica)
California poppy (Eschscholzia californica) Image
Synonyms / Common Names / Related Terms
Eschscholzia californica, Papaveraceae (family).
Mechanism of Action
Pharmacology:
  • Constituents: California poppy yielded alkaloids californidine, escholtzine, N-methyllaurotetanine, caryachine, O-methylcaryachine, and the pavine alkaloid, 6S,12S-neocaryachine-7-O-methyl ether N-metho salt in one study.1
  • Neurological: In an in vitro study, 70% ethanol extract of California poppy was able to bind to 5-HT(1A) and 5-HT(7) receptors.1 The activity on the 5-HT(1A) receptor was at least partly due to the presence of the aporphine alkaloid 3.
Pharmacodynamics/Kinetics:
  • Insufficient available evidence.
References
  1. Gafner, S., Dietz, B. M., McPhail, K. L., Scott, I. M., Glinski, J. A., Russell, F. E., McCollom, M. M., Budzinski, J. W., Foster, B. C., Bergeron, C., Rhyu, M. R., and Bolton, J. L. Alkaloids from Eschscholzia californica and their capacity to inhibit binding of [3H]8-Hydroxy-2-(di-N-propylamino)tetralin to 5-HT1A receptors in Vitro. J Nat Prod  2006;69(3):432-435. 16562853
Licensed by Natural Standard Copyright © 2010 by Natural Standard Corporation. All Rights Reserved.
back to Plant Profiler
back to top