Plant Profiler
Goldenseal (Hydrastis canadensis)
Goldenseal (Hydrastis canadensis) Image
Synonyms / Common Names / Related Terms
Berberine, berberine bisulfate, curcuma, eye balm, eye root, golden root, goldensiegel, goldsiegel, ground raspberry, guldsegl, hydrastis rhizoma, hydrophyllum, Indian dye, Indian paint, Indian plant, Indian turmeric, jaundice root, kanadische gelbwurzel, kurkuma, Ohio curcuma, orange root, Ranunculaceae (family), tumeric root, warnera, wild curcuma, wild turmeric, yellow eye, yellow Indian plant, yellow paint, yellow paint root, yellow puccoon, yellow root, yellow seal, yellow wort.
Note: Goldenseal is sometimes referred to as "Indian turmeric" or "curcuma," but should not be confused with turmeric.
Mechanism of Action
Pharmacology:
  • Constituents: The active ingredients of goldenseal include isoquinoline alkaloids, such as berberine, canadine, and hydrastine. Goldenseal has been reported to contain these alkaloids in the ranges of 1.5-4% hydrastine, 0.5-6% berberine, and 2-3% berberastine.23 Most of the actions of goldenseal have been attributed to hydrastine and berberine. Due to the lack of clinical evidence regarding the use of goldenseal itself, it is unclear whether the actions of its constituents are also attributable to goldenseal preparations.
  • Antibacterial effects: In vitro research assessing the antibacterial activity of berberine has found an aqueous extract containing berberine to have an MIC of 50mcg/mL for Clostridium tetani17, and an MIC <4mcg/mL for Candida krusei26. The MIC for Streptococcus pyogenes was found to be 30mcg/mL; berberine inhibits the adherence of S. pyogenes to epithelial cells possibly by immobilizing fibronectin and hexadecane at concentrations below the MIC.40 Berberine is bactericidal against Vibrio cholera at concentrations of 35mcg/mL, and against Staphylococcus aureus at 50mcg/mL.41 Berberine sulfate has been shown to possess antimicrobial activity against gram positive and gram negative organisms in vitro through inhibition of RNA and protein synthesis.41
  • Antiparasitic effects: Berberine sulfate has been shown to possess antimicrobial activity against protozoal organisms in vitro through inhibition of RNA and protein synthesis.41 In vitro, a methanol extract of berberine has demonstrated parasiticidal activity against T. vaginalis, G. lamblia, and E. histolytica.42 Subsequent in vitro study has found that berberine sulfate (1mg/mL) causes nuclear chromatin clumping in E. histolytica after 24 hours of exposure, irregular shaped vacuoles in G. lamblia after three hours of incubation, and an increased number of autophagic vacuoles in T. vaginalis.43 Berberine (0.5mg) injected into chick embryos reduced the mortality rate of the embryos due to the introduction of trachoma organisms into the yolk sac.11
  • In vitro study has demonstrated the ability of berberine to completely inhibit the growth of promastigotes at a concentration of 5mcg/mL, possibly by inhibiting endogenous respiration of the organism and inhibiting nucleic acid and protein synthesis.44 Subsequent study has shown berberine chloride to interact with Leishmania donovani nuclear DNA, inhibiting the multiplication of amastigotes in macrophage culture in vitro and decreasing parasitic load in animals.16
  • Anti-fungal effects: Berberine, at concentrations of 10mg/mL, has exhibited antifungal activity against Alternaria, Candida albicans, Curvularia, Drchslera, Fusarium, Mucor, and Rhizopus oryzae; concentrations of 25mg/mL have inhibited growth of Aspergillus flavus and Asp. fumigates in vitro.6 Berberine sulfate has been shown to possess antifungal activity in vitro through inhibition of RNA and protein synthesis.41
  • Anti-inflammatory effects: Berberine sulfate administered subcutaneously to the ears of mice in doses of 4-8mg/kg has significantly inhibited xylene-induced swelling.27 Berberine has inhibited edema and inflammation induced in guinea pig paw by carrageenan or zymosan solution.7 Rats treated with 6.6 grams of goldenseal extract in drinking water were repeatedly exposed to a known antigen (keyhole limpet hemocyanin); goldenseal was associated with increased production of IgM following antigen exposure.45 COX-2 regulation has been implicated as a possible mechanism.46,47
  • Anti-proliferative effects: Berberine was shown in vitro to inhibit DNA fragmentation and apoptosis of thymocytes induced by etoposide and camptothecin 48, to affect cell cycle and apoptosis in HeLa/L1210 cells10, and to inhibit synthesis of DNA, RNA, protein, and lipids in ascitic tumor cell lines; however, this inhibition did not carry over to experiments performed in mice49. Berberine significantly inhibited the transformation of lymphocytes, despite the presence of known mitogens in vitro, as measured by [3H]thymidine uptake by lymphocytes.50 After three days of continuous exposure in vitro, berberine significantly inhibited hepatoma cell growth in a dose-dependent manner, and inhibited the release of alpha-fetoprotein after 18 hours of exposure.51 In concentrations of 25mcg/mL, berberine-induced apoptosis during the S-phase of the cell cycle in promyelocytic leukemia HL-60 cells.52 An in vitro study found that 9-ethoxycarbonyl berberine significantly inhibits topoisomerase II.53 Protoberberines are organic cations that are able to intercalate DNA and inhibit topoisomerase I.54 Berberine has also been found to activate macrophages to act against the growth of tumor cells at concentrations above 0.15mcg/mL.28 Additionally, at concentrations above 1.5mcg/mL, berberine has successfully inhibited DNA synthesis in tumor cells. Berberine was shown to induce differentiation of human teratocarcinoma cells into cells with neuronal cell morphology, beginning one day after the addition of 0.1mg/mL berberine to the culture medium.29 In an experiment on mouse skin, berberine inhibited activity of the tumor promoters teleocidin and 12-O-tetradecanoylphorbol-13-acetate.9 In a murine model of Lewis lung carcinoma, oral administration of berberine for two weeks significantly inhibited mediastinal lymph node metastases; however, there was no inhibition of tumor growth in lung parenchyma.55 The addition of berberine to a culture of human brain tumor cell lines resulted in a mean 91% rate of cell death.8 In a rat model of gliosarcoma, berberine administration resulted in a mean 80.9% rate of cell death.8
  • Anti-secretory effects: Berberine sulfate administered orally in doses of 60mg/kg, significantly decreases vascular permeability caused by 0.7% acetic acid in mice.27 Subcutaneous administration of berberine sulfate in doses of 20-50mg/kg decreases vascular permeability induced by histamine.27 Berberine significantly reduces the secretory response of pig jejunal segments to E. coli heat-stable enterotoxin or neostigmine.56,36,57
  • Bilirubin effects: Berberine was shown to increase the secretion of bilirubin in rats with hyperbilirubinemia acutely, although this effect diminished with continued berberine exposure.58 A subsequent study reported berberine to displace bilirubin from albumin both in vitro and in animals, resulting in an increase in serum total and direct bilirubin concentrations.21
  • Cardiovascular effects: A 30-minute infusion of berberine in 12 patients with congestive heart failure at a rate of 0.2mg/kg has been shown to significantly improve systemic and pulmonary vascular resistance, right atrial and left ventricular end-diastolic pressures, cardiac index, and left ventricular ejection fraction.18 Intravenous berberine at a concentration of 0.2mg/kg/min significantly raised left ventricular end-diastolic pressure in anesthetized dogs with embolized left main coronary arteries.59 Berberine has exhibited positive inotropic effects in dogs and prevents/reverses ouabain-induced ventricular arrhythmias.2,3 Berberine plasma concentrations greater than 0.11mg/L have been associated with a significant decrease in the occurrence of ventricular premature beats and a significant increase in left ventricular ejection fraction in patients with congestive heart failure vs. plasma concentrations less than 0.11mg/L.60 Animal experiments have reported berberine to restore ventricular arrhythmias and atrial fibrillation to normal sinus rhythm.4 Berberine 0.2-0.7mg/kg/min increases cardiac output and decreases total peripheral resistance and heart rate in animals; doses of 0.02mg/kg/min only increase cardiac output.61 Berberine sulfate bolus injection (1mg/kg), administered to rats one minute after undergoing coronary artery occlusion significantly reduced early mortality from ventricular fibrillation or complete atrioventricular block (36% mortality vs. 66% mortality in a control group).5 Berberine sulfate solution (5mg/mL) produced a dose-dependent decrease in blood pressure in anesthetized dogs, cats, frogs, and rats that was not inhibited by intravenous atropine, mepyramine maleate, pentolinium tartrate, propranolol or phenoxybenzamine.22 Intravenous berberine caused a significant decrease in systolic and diastolic blood pressure in rats at doses of 2-8mg/kg.24 Berberine has caused bradycardia in isolated right and left atria excised from guinea pigs, which was not reversible by atropine.35 An alcoholic extract of goldenseal has been shown in vitro to cause dose-dependent inhibition of epinephrine, serotonin, and histamine-induced aortic contraction.25 Dose-dependent vasoconstriction was observed beginning at a concentration of 50mcL/mL of goldenseal, although neither berberine nor hydrastine alone demonstrated this effect. Extracts of berberine alone showed inhibitory activity on adrenaline-induced aortic contraction. Hydrastine alone was ineffective. Berberine has been found to competitively inhibit the binding of yohimbine in a fashion similar to that of clonidine, suggesting that berberine may possess partial agonist activity at platelet alpha-2 receptors.39
  • Coagulation effects: The effects of goldenseal on coagulation are not clear. In vitro study has found that berberine inhibits platelet-activating factor and aggregation of platelets with a reported 50% inhibition at a concentration of 38mcg/mL, and inhibits the binding of platelet-activating factor to rabbit platelets with 50% inhibition seen at a concentration of 480mcg/mL.19 Other animal research has reported that berberine inhibits platelet aggregation caused by ADP, arachidonic acid, and collagen, and decreases thromboxane-B2 in rats with ischemic cerebral artery occlusion at doses of 20mg/kg for 1-5 days.20 It has also been noted that goldenseal may reduce the anticoagulant effects of warfarin due to antagonist properties.
  • Cytochrome P450 effects: Goldenseal has been found to inhibit cytochrome P450 3A (4,5) and 2D6 enzyme.33,34
  • CNS effects: In animals, berberine produced sedation and potentiated the sedative effects of pentobarbitone when administered via intraperitoneal or intraventricular routes.37 Berberine has been shown to reduce spontaneous motor activity and prolong hexobarbitone-induced sleeping time when administered to mice.22 The administration of berberine (0.1-0.5g/kg) for 14 days was effective in improving scopolamine-induced amnesia in rats, an effect that was augmented by physostigmine and neostigmine.38
  • Gastrointestinal effects: Oral berberine sulfate (40-80mg/kg) has significantly decreased the occurrence of diarrhea induced by ingestion of castor oil and Cassia angustifolia in mice.27 In 20 healthy subjects, the oral administration of 1.2g of berberine significantly delayed small intestinal transit time of a meglucamine diatrizoate and sorbitol test mixture (71.10 ±22.04 minutes in control vs. 98.25 ±29.03 minutes with berberine, p<0.01).62 In animal study, berberine significantly potentiated emesis in dogs.63
  • Endocrine effects: Berberine was reported to improve insulin resistance and liver glycogen levels similarly to metformin in rats fed a high-fat diet.12 Diabetic rats treated with berberine had significantly lower blood sugar concentrations than control rats.13 Goldenseal has also reduced hyperphagia and polydipsia associated with streptozotocin-induced diabetes mellitus in animals.14 In vitro study reports that berberine decreased glucose absorption by Caco-2 cells.15
  • Berberine, an active component of goldenseal, has been shown to inhibit parathyroid hormone-stimulated bone resorption in animal study.64 Berberine, in doses of 30-50mg/kg daily, has demonstrated an ability to prevent a decrease in bone mineral density of lumbar vertebra in ovariectomized rats and to induce apoptosis of osteoclastic cells.64
  • Muscle relaxant effects: Goldenseal extract, at a cumulative dose of 5mcg/mL, caused complete relaxation of carbachol-precontracted guinea pig trachea in vitro and a significant increase in cAMP levels.65 Berberine sulphate (20mcg/mL) pretreatment blocked the response of ileum, trachea, and rectal muscles to acetylcholine in animals.1 Similarly, an ethanol extract of goldenseal induced relaxation in rabbit bladder muscle in vitro.66 This relaxation was partly blocked by the addition of propranolol to the medium, suggesting a mechanism partially mediated through β-adrenoreceptors. An alcoholic extract of goldenseal inhibited acetylcholine, oxytocin, and serotonin-induced contractions of rat uterus cells in a dose dependent fashion in vitro.30 Berberine inhibited muscle contractions of guinea pig ileum and rat uterus induced by acetylcholine, carbachol, histamine, potassium chloride, and bradykinin.22 Berberine increased the amplitude of slow-response action potentials induced by histamine by 6.2%, increased the maximum rate of depolarization by 21.1%, increased the action potential duration (APD) by 50.1% (APD 50) and 47.2% (APD 100), and effective refractory period by 92.2%.31
Pharmacodynamics/Kinetics:
  • Goldenseal is traditionally believed to be poorly absorbed from the gastrointestinal tract.
  • Berberine plasma concentrations greater than 0.11mg/L have been associated with a significant decrease in the occurrence of ventricular premature beats and a significant increase in left ventricular ejection fraction in patients with congestive heart failure vs. plasma concentrations less than 0.11mg/L.60
  • In vitro, goldenseal extract has been shown to weakly inhibit CYP 3A432; however, this interaction has not been sufficiently evaluated in humans.
References
  1. Kulkarni, S. K., Dandiya, P. C., and Varandani, N. L. Pharmacological investigations of berberine sulphate. Jpn J Pharmacol  1972;22(1):11-16. 4260852
  2. Krol R, Zalewski A, Cheung W, and et al. Additive effects of berberine and ouabain on myocardial contractility. Clin Res 1982;30(3):673A.
  3. Krol R, Zalewski A, and Maroko PR. Beneficial effects of berberine, a new positive inotropic agent, on digitalis-induced ventricular arrhythmias. Circulation 1982;66(suppl 2):56.
  4. Ksiezycka E, Cheung W, and Maroko PR. Antiarrhythmic effects of berberine on aconitine-induced ventricular and supraventricular arrhythmias. Clinical Research 1983;31(2):197A.
  5. Ribeiro LG, Bowker BL, Maroko PR, and et al. Beneficial effects of berberine on early mortality after experimental coronary artery occlusion in rats. Circulation 1982;66(II):56.
  6. Mahajan VM, Sharma A, and Rattan A. Antimycotic activity of berberine sulphate: an alkaloid from an Indian medicinal herb. Sabouraudia 1982;20:79-81.
  7. Ivanovska N and Philipov S. Study on the anti-inflammatory action of Berberis vulgaris root extract, alkaloid fractions and pure alkaloids. Int J Immunopharmac 1996;18(10):553-561.
  8. Zhang, R. X., Dougherty, D. V., and Rosenblum, M. L. Laboratory studies of berberine used alone and in combination with 1,3-bis(2-chloroethyl)-1-nitrosourea to treat malignant brain tumors. Chin Med J (Engl) 1990;103(8):658-665. 2122945
  9. Nishino H, Kitagawa K, Fujiki H, and et al. Berberine sulfate inhibits tumor-promoting activity of teleocidin in two-stage carcinogenesis on mouse skin. Oncology 1986;43:131-134.
  10. Jantova, S., Cipak, L., Cernakova, M., and Kost'alova, D. Effect of berberine on proliferation, cell cycle and apoptosis in HeLa and L1210 cells. J Pharm Pharmacol 2003;55(8):1143-1149. 12956905
  11. Sabir M, Mahajan VM, Mohapatra LN, and et al. Experimental study of the antitrachoma action of berberine. Indian J Med Res 1976;64(8):1160-1167. 992845
  12. Gao, C. R., Zhang, J. Q., and Huang, Q. L. [Experimental study on berberin raised insulin sensitivity in insulin resistance rat models]. Zhongguo Zhong Xi Yi Jie He Za Zhi  1997;17(3):162-164. 9863084
  13. Ni, Y. X. [Therapeutic effect of berberine on 60 patients with type II diabetes mellitus and experimental research]. Zhong Xi Yi Jie He Za Zhi - Chinese Journal of Modern Developments in Traditional Medicine 1988;8(12):711-3, 707. 3248329
  14. Swanston-Flatt SK, Day C, Bailey CJ, and et al. Evaluation of traditional plant treatments for diabetes: studies in streptozotocin diabetic mice. Acta Diabetol Lat 1989;26:51-55.
  15. Pan, G. Y., Huang, Z. J., Wang, G. J., Fawcett, J. P., Liu, X. D., Zhao, X. C., Sun, J. G., and Xie, Y. Y. The antihyperglycaemic activity of berberine arises from a decrease of glucose absorption. Planta Med 2003;69(7):632-636. 12898419
  16. Ghosh AK, Bhattacharyya FK, and Ghosh DK. Leishmania donovani: amastigote inhibition and mode of action of berberine. Experimental Parasitology 1985;60:404-413.
  17. Palasuntheram C, Iyer KS, de Silva LB, and et al. Antibacterial activity of Coscinium fenestratum Colebr against Clostridium tetani. Ind J Med Res 1982;76(Suppl):71-76.
  18. Marin-Neto, J. A., Maciel, B. C., Secches, A. L., and Gallo, Junior L. Cardiovascular effects of berberine in patients with severe congestive heart failure. Clin Cardiol  1988;11(4):253-260. 3365876
  19. Tripathi YB and Shukla SD. Berberis artistata inhibits PAF induced aggregation of rabbit platelets. Phytotherapy Research 1996;10:628-630.
  20. Wu, J. F. and Liu, T. P. [Effects of berberine on platelet aggregation and plasma levels of TXB2 and 6-keto-PGF1 alpha in rats with reversible middle cerebral artery occlusion]. Yao Xue Xue Bao  1995;30(2):98-102. 7785438
  21. Chan, E. Displacement of bilirubin from albumin by berberine. Biol Neonate 1993;63(4):201-208. 8513024
  22. Sabir M and Bhide NK. Study of some pharmacological actions of berberine. Ind J Physiol & Pharmac 1971;15(3):111-132.
  23. Hamon, NW. Goldenseal  CPJ-RPC 1990;508-510.
  24. Chun YT, Yip TT, Lau KL, and et al. A biochemical study on the hypotensive effect of berberine in rats. Gen Pharmac 1979;10:177-182. 572797
  25. Palmery M, Leone M, Pimpinella G, and et al. Effects of Hydrastis canadensis L. and the two major alkaloids berberine and hydrastine on rabbit aorta. Pharmacological Research 1993;27(suppl 1):73-74.
  26. Park, K. S., Kang, K. C., Kim, J. H., Adams, D. J., Johng, T. N., and Paik, Y. K. Differential inhibitory effects of protoberberines on sterol and chitin biosyntheses in Candida albicans. J Antimicrob Chemother 1999;43(5):667-674. 10382888
  27. Zhang, M. F. and Shen, Y. Q. [Antidiarrheal and anti-inflammatory effects of berberine]. Zhongguo Yao Li Xue Bao  1989;10(2):174-176. 2816420
  28. Kumazawa Y, Itagaki A, Fukumoto M, and et al. Activation of peritoneal macrophages by berberine-type alkaloids in terms of induction of cytostatic activity. Int J Immunopharmac 1984;6(6):587-592.
  29. Chang, K. S., Gao, C., and Wang, L. C. Berberine-induced morphologic differentiation and down-regulation of c- Ki-ras2 protooncogene expression in human teratocarcinoma cells. Cancer Lett  12-3-1990;55(2):103-108. 2265407
  30. Cometa MF, Abdel-Haq H, and Palmery M. Spasmolytic activities of Hydrastis canadensis L. on rat uterus and guinea-pig trachea. Phytotherapy Research 1998;12(suppl 1):S83-S85.
  31. Huang W, Zhang Z, and Xu Y. Study of the effects and mechanisms of berberine on slow-response action potentials. Journal of Electrocardiology 1990;23(3):231-234.
  32. Budzinski, J. W., Foster, B. C., Vandenhoek, S., and Arnason, J. T. An in vitro evaluation of human cytochrome P450 3A4 inhibition by selected commercial herbal extracts and tinctures. Phytomedicine 2000;7(4):273-282. 10969720
  33. Gurley, B. J., Swain, A., Hubbard, M. A., Hartsfield, F., Thaden, J., Williams, D. K., Gentry, W. B., and Tong, Y. Supplementation with goldenseal (Hydrastis canadensis), but not kava kava (Piper methysticum), inhibits human CYP3A activity in vivo. Clin Pharmacol Ther 2008;83(1):61-69. 17495878
  34. Gurley, B. J., Gardner, S. F., Hubbard, M. A., Williams, D. K., Gentry, W. B., Khan, I. A., and Shah, A. In vivo effects of goldenseal, kava kava, black cohosh, and valerian on human cytochrome P450 1A2, 2D6, 2E1, and 3A4/5 phenotypes. Clin Pharmacol Ther  2005;77(5):415-426. 15900287
  35. Shaffer, J. E. Inotropic and chronotropic activity of berberine on isolated guinea pig atria. J Cardiovasc Pharmacol 1985;7(2):307-315. 2581085
  36. Zhu, B. and Ahrens, F. Antisecretory effects of berberine with morphine, clonidine, L- phenylephrine, yohimbine or neostigmine in pig jejunum. Eur J Pharmacol 12-9-1983;96(1-2):11-19. 6363101
  37. Shanbhag, S. M., Kulkarni, H. J., and Gaitonde, B. B. Pharmacological actions of berberine on the central nervous system. Jpn J Pharmacol 1970;20(4):482-487. 5312930
  38. Peng, W. H., Hsieh, M. T., and Wu, C. R. Effect of long-term administration of berberine on scopolamine-induced amnesia in rats. Jpn J Pharmacol 1997;74(3):261-266. 9268086
  39. Hui, K. K., Yu, J. L., Chan, W. F., and Tse, E. Interaction of berberine with human platelet alpha 2 adrenoceptors. Life Sci  1991;49(4):315-324. 1649363
  40. Sun D, Courtney HS, and Beachey EH. Berberine sulfate blocks adherence of Streptococcus pyogenes to epithelial cells, fibronectin, and hexadecane. Antimicrobial Agents and Chemotherapy 1988;32(9):1370-1374.
  41. Amin, A. H., Subbaiah, T. V., and Abbasi, K. M. Berberine sulfate: antimicrobial activity, bioassay, and mode of action. Can J Microbiol  1969;15(9):1067-1076. 4906191
  42. Kaneda Y, Tanaka T, and Saw T. Effects of berberine, a plant alkaloid, on the growth of anaerobic protozoa in axenic culture. Tokai J Exp Clin Med 1990;15(6):417-423.
  43. Kaneda Y, Torii M, Tanaka T, and et al. In vitro effects of berberine sulphate on the growth and structure of Entamoeba histolytica, Giardia lamblia and Trichomonas vaginalis. Annals of Tropical Medicine and Parasitology 1991;85(4):417-425.
  44. Ghosh, A. K., Rakshit, M. M., and Ghosh, D. K. Effect of berberine chloride on Leishmania donovani. Indian J Med Res  1983;78:407-416. 6674161
  45. Rehman J, Dillow JM, Carter SM, and et al. Increased production of antigen-specific immunoglobulins G and M following in vivo treatment with the medicinal plants Echinacea angustifolia and Hydrastis canadensis. Immunology Letters 1999;68:391-395.
  46. Kuo, C. L., Chi, C. W., and Liu, T. Y. The anti-inflammatory potential of berberine in vitro and in vivo. Cancer Lett 1-28-2004;203(2):127-137. 14732220
  47. Lee, D. U., Kang, Y. J., Park, M. K., Lee, Y. S., Seo, H. G., Kim, T. S., Kim, C. H., and Chang, K. C. Effects of 13-alkyl-substituted berberine alkaloids on the expression of COX-II, TNF-alpha, iNOS, and IL-12 production in LPS-stimulated macrophages. Life Sci  8-1-2003;73(11):1401-1412. 12850501
  48. Miura N, Yamamoto M, Ueki T, and et al. Inhibition of thymocyte apoptosis by berberine. Biochemical Pharmacology 1997;53:1315-1322.
  49. Creasey, W. A. Biochemical effects of berberine. Biochem Pharmacol  4-1-1979;28(7):1081-1084. 444265
  50. Ckless, K., Schlottfeldt, J. L., Pasqual, M., Moyna, P., Henriques, J. A., and Wajner, M. Inhibition of in-vitro lymphocyte transformation by the isoquinoline alkaloid berberine. J Pharm Pharmacol  1995;47(12A):1029-1031. 8932689
  51. Chi, C. W., Chang, Y. F., Chao, T. W., Chiang, S. H., P'eng, F. K., Lui, W. Y., and Liu, T. Y. Flowcytometric analysis of the effect of berberine on the expression of glucocorticoid receptors in human hepatoma HepG2 cells. Life Sci  1994;54(26):2099-2107. 7516035
  52. Kuo CL, Chou CC, and Yung BY. Berberine complexes with DNA in the berberine-induced apoptosis in human leukemic HL-60 cells. Cancer Letters 1995;93:193-200.
  53. Krishnan, P. and Bastow, K. F. The 9-position in berberine analogs is an important determinant of DNA topoisomerase II inhibition. Anticancer Drug Des 2000;15(4):255-264. 11200501
  54. Li, T. K., Bathory, E., LaVoie, E. J., Srinivasan, A. R., Olson, W. K., Sauers, R. R., Liu, L. F., and Pilch, D. S. Human topoisomerase I poisoning by protoberberines: potential roles for both drug-DNA and drug-enzyme interactions. Biochemistry 6-20-2000;39(24):7107-7116. 10852708
  55. Mitani, N., Murakami, K., Yamaura, T., Ikeda, T., and Saiki, I. Inhibitory effect of berberine on the mediastinal lymph node metastasis produced by orthotopic implantation of Lewis lung carcinoma. Cancer Lett  4-10-2001;165(1):35-42. 11248416
  56. Zhu B and Ahrens FA. Effect of berberine on intestinal secretion mediated by Escherichia coli heat-stable enterotoxin in jejunum of pigs. Am J Vet Res 1982;43(9):1594-1598.
  57. Khin, Maung U. and Nwe, Nwe Wai. Effect of berberine on enterotoxin-induced intestinal fluid accumulation in rats. J Diarrhoeal Dis Res 1992;10(4):201-204. 1296936
  58. Chan, M. Y. The effect of berberine on bilirubin excretion in the rat. Comp Med East West 1977;5(2):161-168. 415839
  59. Vik-Mo, H, Faria DB, Cheung W, and et al. Beneficial effects of berberine on left ventricular function in dogs with heart failure. Clinical Research 1983;31(2):224a.
  60. Zeng, X. and Zeng, X. Relationship between the clinical effects of berberine on severe congestive heart failure and its concentration in plasma studied by HPLC. Biomed Chromatogr 1999;13(7):442-444. 10534753
  61. Zalewski A, Krol R, and Maroko PR. Berberine, a new inotropic agent - distinction between its cardiac and peripheral responses. Clin Res 1983;31(2):227A.
  62. Yuan, J., Shen, X. Z., and Zhu, X. S. [Effect of berberine on transit time of human small intestine]. Zhongguo Zhong Xi Yi Jie He Za Zhi  1994;14(12):718-720. 7719104
  63. Sabir M, Akhter MH, and Bhide NK. Further studies on pharmacology of berberine. Ind J Physiol Pharmac 1978;22(1):9-23.
  64. Li, H., Miyahara, T., Tezuka, Y., Namba, T., Suzuki, T., Dowaki, R., Watanabe, M., Nemoto, N., Tonami, S., Seto, H., and Kadota, S. The effect of kampo formulae on bone resorption in vitro and in vivo. II. Detailed study of berberine. Biol Pharm Bull 1999;22(4):391-396. 10328560
  65. Abdel-Haq, H., Cometa, M. F., Palmery, M., Leone, M. G., Silvestrini, B., and Saso, L. Relaxant effects of Hydrastis canadensis L. and its major alkaloids on guinea pig isolated trachea. Pharmacol Toxicol  2000;87(5):218-222. 11129501
  66. Bolle P, Cometa MF, Palmery M, and et al. Response of rabbit detrusor muscle to total extract and major alkaloids of Hydrastis canadensis. Phytotherapy Research 1998;12:S86-S88.
Licensed by Natural Standard Copyright © 2010 by Natural Standard Corporation. All Rights Reserved.
back to Plant Profiler
back to top