|
Plant Profiler
Populus (Populus tremuloides)
|
|
|
Synonyms / Common Names / Related Terms
Aspen, balm of gilead, cottonwood, pando, poplar, popple, Populus sieboldii, Populus tremula L., Populus tremuloides, quaking aspen, Salicaceae (family), the trembling giant.
Combination product example: Phytodolor N (Populus tremula, Solidago virgaurea, Fraxinus excelsior).
Mechanism of Action
Pharmacology:
- Constituents: The water extract of Populus sieboldii Miquel contains pyrocatechol and salicyl alcohol.2
- Antibacterial effects: In an in vitro study, extracts from aspen bark (Populus tremula L.) with the use of petroleum ether (lipids I) and diethyl ether (lipids II) destroyed all Streptococcus pneumoniae and Haemophilus influenzae cells.1 The minimum inhibiting concentration (MIC) for Streptococcus pneumoniae was 0.5-50mg/mL for lipids I and 0.0005-0.5mg/mL for lipids II. The MIC for Haemophilus influenzae was 30-50mg/mL.
- Antiplatelet effects: In an in vitro study, the water extract of Populus sieboldii Miquel inhibited arachidonic acid-induced platelet aggregation.2 The active constituents pyrocatechol and salicyl alcohol were isolated from this extract and pyrocatechol inhibited platelet aggregation induced by arachidonic acid with an IC100 value of 4mcM, which was 25 times more potent than aspirin.
Pharmacodynamics/Kinetics:
- In an in vitro study, extracts from aspen bark (Populus tremula L.) with the use of petroleum ether (lipids I) and diethyl ether (lipids II) destroyed all Streptococcus pneumoniae and Haemophilus influenzae cells.1 The minimum inhibiting concentration (MIC) for Streptococcus pneumoniae was 0.5-50mg/mL for lipids I and 0.0005-0.5mg/mL for lipids II. The MIC for Haemophilus influenzae was 30-50mg/mL.
References
- Faustova, N. M., Faustova, M. E., and Deineko, I. P. [Antibacterial activity of aspen bark extracts against some pneumotropic microorganisms]. Zh Mikrobiol Epidemiol Immunobiol 2006;(3):3-7. 16830580
- Kagawa, K., Tokura, K., Uchida, K., Kakushi, H., and Shike, T. Platelet aggregation inhibitors from Populus sieboldii Miquel. Chem Pharm Bull (Tokyo) 1992;40(8):2191-2192. 1423779
Licensed by Natural Standard Copyright © 2010 by Natural Standard Corporation. All Rights Reserved.
back to Plant Profiler back to top
|