|
Plant Profiler
Kudzu (Pueraria lobata)
|
|
|
Synonyms / Common Names / Related Terms
Arrowroot, biochanin A, daidzein, daidzein 8-C-glucoside, daidzin, Fabaceae (family), flos puerariae, ge-gen, gegen-tanj (TJ-1), genistein, genistin, glycitin, Japanese arrowroot, kaikasaponin III (KS-III), kakkonto, Kampo, kudzu root, Kwao Kruea Khao, Leguminosae (family), NPI-028, NPI-031, NPI-031G, pedunsaponin B2, pedunsaponin C3, puer, Pueraria lobata, Pueraria lobata L., Pueraria lobata Ohwi, Pueraria lobata root decoction, Pueraria lobata (Willd.), Pueraria lobata (Willd.) Ohwi, Pueraria mirifica, Pueraria montana, Pueraria omeiensis, Pueraria peduncularis, Pueraria phaseoloides, Pueraria thomsonii, Pueraria thunbergiana, Puerariae flos, Puerariae radix, puerariae surculus, puerariae flos, puerarin, radix puerariae, spinasterol, tectoridin, tectorigenin, Tianbaokang, Yufengningxin.
Mechanism of Action
Pharmacology:
- Constituents: The active constituents of kudzu include daidzin, daidzein, puerarin, genistin, genistein, tectorigenin, glycitin, tectoridin, 6"-O-xylosyltectoridin, 6"-O-xyloglycitin, biochanin A, and spinasterol.42,48,49,50,51,52,53,54,5
- Two oleanane-type triterpene saponins named pedunsaponins B2 and C3 were isolated from the roots of Pueraria peduncularis.55 Their structures were determined to be 3-O-(6-O-methyl)-beta-glucuronopyranosyl-3beta,15alpha-dihydroxyolean-12-en-16-one (2), and 3-O-beta-glucopyranosyl-(1 --> 3)-beta-glucuronopyranosyl-3beta,15alpha-dihydroxyoleana-12-en-16-one (3).
- Alcohol effects: The isoflavones daidzein, genistein, formononetin and biochanin A isolated from kudzu root have shown to be potent reversible human alcohol dehydrogenase (ADH) isoenzyme inhibitors in vitro. The isoflavones produced a competitive inhibition of gamma(g)2-g2-ADH-isoenzyme with respect to ethanol and an uncompetitive inhibition of gamma(g)2-g2-ADH-isoenzyme with respect to NAD+. The most potent inhibitor was genistein with an effective concentration of 0.1micromole. Although flavones such as apigenin, kaempferol, 7-hydroxyflavone, and galangin also demonstrated ADH-inhibiting activity, the effects were more pronounced with isoflavones. No effects were seen in B1B1-ADH isoenzymes and B2B2-ADH isoenzymes when concentrations up to 20micromoles were used.41
- The isoflavones in kudzu root extract may suppress alcohol intake and alcohol withdrawal symptoms in animals, although the mechanism is unclear.42,56,39,57
- Kudzu may decrease peak blood alcohol levels due to delayed gastric emptying, exposing alcohol to a longer time for first-pass metabolism in the stomach.43,44 Slowed gastric emptying may prolong the effects of alcohol.
- Kudzu may contribute to alleviating the adverse effects of ethanol ingestion by enhancing the lipid metabolism as well as the hepatic antioxidant defense system.58 Kudzu may also have an appetite suppressant effect for alcohol.59
- The flowers of kudzu exhibit protective effects against ethanol-induced apoptosis in human neuroblastoma cells by inhibiting the expression of a protease, caspase-3 that is responsible for proteolytic cleavage of many proteins.60
- Anti-bacterial effects: Arrowroot tea (Puerariae radix) inhibited microbial growth of both Gram-negative and Gram-positive food borne pathogens in various foods, especially liquid foods.61
- Anti-cancer effects: Genistein, biochanin A and daidzein appear to have cytotoxic activity.62,51 Preliminary research results suggest that biochanin A and genistein inhibit the cell growth of stomach cancer cell lines in vitro through activation of a signal transduction pathway for apoptosis.63 The constituent tectorigenin demonstrated antiproliferative activity against human cancer (HL-60) cells by the induction of differentiation in the cells and a reduction in the expression of Bcl-2, an antiapoptotic protein.51
- Anti-inflammatory effects: Kudzu has the ability to decrease prostaglandin E2 and tumor necrosis factor (TNF)-alpha release, both of which are involved in inflammatory process.1
- Antioxidant effects: Kudzu and its constituents may have antioxidant activity.64,40,2,3 Pueraria lobata has shown more potent antioxidant activity than that of Pueraria thomsonii likely due to its higher contents of isoflavonoids.65
- Puerarin may protect neurons against oxidative stress.66 It appears to block apoptosis in its early stages via the regulation of anti- and pro-apoptotic proteins, as well as by the attenuation of caspase-3 activation in H2O2-induced PC12 cells.
- Puerarin promoted the activity of super oxide dismutase in the brain, liver, and serum in animal studies.67,68
- Anti-thrombotic effects: In clinical and animal studies, Pueraria lobata has displayed antithrombotic activity.19,20,21,22,23,24,18,25,26 It may inhibit the increase of platelet aggregation and blood viscosity.22,24,25,26 Puerarin can increase superoxide dismutase activity, decrease lipoprotein level and enhance the activity of fibrinolysis.19
- Cardiovascular effects: Early preclinical and animal research suggests kudzu has a protective effect against myocardial ischemia and may increase cardiac function.13,69,70,22,4,71,72,73,11,29,74 Animal research suggests that puerarin may reduce both systolic and diastolic blood pressure, and diminish myocardial oxygen consumption.37
- The kudzu constituent, daidzein, may have antiarrhythmic properties.35,36,37,38 These effects may be related to its inhibition of Na+ or Ca2+ influx and its blocking beta-adrenergic receptor.75,76,71,36,37
- Kudzu increased nitric oxide content in heart, brain, liver, and kidney.77,78
- Puerarin may have vasorelaxant properties, possibly by blocking beta-adrenergic receptors.15,28,45
- Puerarin decreases plasma renin and angiotensin II activity.27
- Cytochrome P450 system effects: Early research suggests that kudzu inhibits and induces cytochrome P450 isoenzymes; however, it is unclear which CYP isoenzymes are affected and to what degree.40
- Estrogenic effects: Isoflavone constituents have both estrogenic and antiestrogenic activity.79,80,81,31,32,46,82,16,47,48,33,34
- Based on lab and animal research, puerarin, genistin, glycetein, and daidzein are considered phytoestrogens.83,80,82 These phytoestrogens may have additive or synergistic effects with each other.12,33
- Based on an open label trial, Pueraria mirifica is thought to be involved in alleviating symptoms such as hot flashes and night sweats in perimenopausal women17 and affecting cognitive function in postmenopausal women.
- Hepatic effects: Puerarin showed potent hepatoprotective activity, but did not inhibit beta-glucuronidase; however, daidzein, which is produced by human intestinal bacteria, potently inhibited beta-glucuronidase.6,7
- Hypoglycemic effects: Based on animal studies, kudzu and its constituents may have hypoglycemic effects.8,9,10 Puerarin has increased glucose utilization to lower plasma glucose.84 It has also activated alpha1-adrenoceptors on the adrenal gland to enhance the secretion of beta-endorphin to result in a decrease of plasma glucose.85,30
- Compounds that target the peroxisome proliferator-activated receptors PPARalpha and PPARgamma are used to correct dyslipidemia and to restore glycemic balance, respectively.86 Because the majority of diabetic patients suffer from atherogenic lipid abnormalities in addition to insulin resistance, ligands are required that can activate both PPARalpha and PPARgamma. Pueraria thomsonii significantly activated PPARalpha and PPARgamma and bioassay-guided fractionation isolated daidzein as the PPAR-activating compound. However, daidzein was not as potent as other common isoflavones.
- Neurologic effects: Kudzu may provide protection for neurons.87,14,88
- Daidzin may act by inhibiting serotonin and dopamine metabolism.89
- Puerarin showed an improvement effect against the memory impairment in the modeling aging-mice induced by D-galactose.68
- Based on animal studies, puerarin reduced flow velocities of both the middle cerebral artery and anterior cerebral artery; cerebral blood flow was enhanced due to dilatation of the intracranial arteries.90,91
- Osteoporotic effects: In animal studies, Pueraria lobata increased bone mineral density.34 Puerarin has also suppressed bone resorption and promoted bone formation in animal studies.14,46
Pharmacodynamics/Kinetics:
- Absorption: Puerarin is rapidly, but incompletely, absorbed from the gastrointestinal tract.52,92
- Bioavailability: Plasma daidzin concentration-time curves determined the crude extract daidzin has approximately 10 times greater bioavailability than the pure compound based on an animal study.50
- Experimental results demonstrated that the excretive amounts of puerarin reached its maximum three hours after ingesting puerarin.93
- Distribution: Puerarin is distributed widely in the body.92
- Metabolism: When puerarin or daidzin were incubated for 24 hours with human intestinal bacteria, two metabolites, daidzein and calycosin, were produced from them, respectively.94 The metabolic time course of puerarin was as follows: at an early time, puerarin was converted to daidzin, and then calycosin. The metabolic time course of daidzin by human intestinal bacteria was also similar to that of puerarin. Puerarin and daidzin were transformed to their aglycones by the bacteria producing beta-glucuronidase, C-glycosidase and beta-glycosidase, respectively.21
- Kudzu may decrease peak blood alcohol levels due to delayed gastric emptying, exposing alcohol to a longer time for first-pass metabolism in the stomach.43,44
- Elimination: Puerarin is eliminated at a fairly rapid rate. The blood level of puerarin following intravenous administration in rats was found to decrease in the elimination phase with a half-life of 18 minutes.92
References
- Kim, I. T., Park, Y. M., Shin, K. M., Ha, J., Choi, J., Jung, H. J., Park, H. J., and Lee, K. T. Anti-inflammatory and anti-nociceptive effects of the extract from Kalopanax pictus, Pueraria thunbergiana and Rhus verniciflua. J Ethnopharmacol 2004;94(1):165-173. 15261979
- Lau, C. S., Carrier, D. J., Beitle, R. R., Howard, L. R., Lay, J. O., Liyanage, R., and Clausen, E. C. A glycoside flavonoid in Kudzu (Pueraria lobata): identification, quantification, and determination of antioxidant activity. Appl Biochem Biotechnol 2005;121-124:783-794. 15930558
- Zhang, G. and Fang, S. [Antioxidation of Pueraria lobata isoflavones (PLIs)]. Zhong Yao Cai 1997;20(7):358-360. 12572435
- Liu, Q., Wang, L., Lu, Z., Li, S., and Xiong, Y. [Effect of puerarin on coronary collateral circulation in dogs with experimental acute myocardial infarction]. Zhongguo Zhong Yao Za Zhi 1999;24(5):304-6, 320. 12205891
- Zhang, S., Ji, G., and Liu, J. Reversal of chemical-induced liver fibrosis in Wistar rats by puerarin. J Nutr Biochem 10-10-2005;16426832
- Bae, H. S., Kim, Y. S., Cho, K. H., Lee, K. S., Kim, J. J., Lee, H. U., and Kim, D. H. Hepatoprotective activity of reduohanxiao-tang (yuldahanso-tang) is related to the inhibition of beta-glucuronidase. Am J Chin Med 2003;31(1):111-117. 12723760
- Lee, H. W., Choo, M. K., Bae, E. A., and Kim, D. H. Beta-glucuronidase inhibitor tectorigenin isolated from the flower of Pueraria thunbergiana protects carbon tetrachloride-induced liver injury. Liver Int 2003;23(4):221-226. 12895260
- Choi, J., Shin, M. H., Park, K. Y., Lee, K. T., Jung, H. J., Lee, M. S., and Park, H. J. Effect of kaikasaponin III obtained from Pueraria thunbergiana flowers on serum and hepatic lipid peroxides and tissue factor activity in the streptozotocin-induced diabetic rat. J Med Food 2004;7(1):31-37. 15117550
- Duan, H. J., Liu, S. X., Zhang, Y. J., Liu, Q. J., He, N., and Li, Y. M. [Effects of puerarin on renal function, expressions of MMP-2 and TIMP-2 in diabetic rats]. Yao Xue Xue Bao 2004;39(7):481-485. 15493832
- Song, C. Y. and Bi, H. M. [Effects of puerarin on plasma membrane GLUT4 content in skeletal muscle from insulin-resistant Sprague-Dawley rats under insulin stimulation]. Zhongguo Zhong Yao Za Zhi 2004;29(2):172-175. 15719689
- Xuan, B., Zhou, Y. H., Yang, R. L., Li, N., Min, Z. D., and Chiou, G. C. Improvement of ocular blood flow and retinal functions with puerarin analogs. J Ocul Pharmacol Ther 1999;15(3):207-216. 10385129
- Trisomboon, H., Malaivijitnond, S., Watanabe, G., and Taya, K. Ovulation block by Pueraria mirifica: a study of its endocrinological effect in female monkeys. Endocrine 2005;26(1):33-39. 15805583
- Fan, L. L., Sun, L. H., Li, J., Yue, X. H., Yu, H. X., and Wang, S. Y. The protective effect of puerarin against myocardial reperfusion injury. Study on cardiac function. Chin Med J (Engl) 1992;105(1):11-17. 1576864
- Li, X., Sun, S., and Tong, E. Experimental study on the protective effect of puerarin to Parkinson disease. J Huazhong Univ Sci.Technolog Med Sci 2003;23(2):148-150. 12973933
- Dong, K., Tao, Q. M., Xia, Q., Shan, Q. X., and Pan, G. B. [Endothelium-independent vasorelaxant effect of puerarin on rat thoracic aorta]. Zhongguo Zhong Yao Za Zhi 2004;29(10):981-984. 15631089
- Woo, J., Lau, E., Ho, S. C., Cheng, F., Chan, C., Chan, A. S., Haines, C. J., Chan, T. Y., Li, M., and Sham, A. Comparison of Pueraria lobata with hormone replacement therapy in treating the adverse health consequences of menopause. Menopause 2003;10(4):352-361. 12851519
- Lamlertkittikul, S. and Chandeying, V. Efficacy and safety of Pueraria mirifica (Kwao Kruea Khao) for the treatment of vasomotor symptoms in perimenopausal women: Phase II Study. J Med Assoc Thai 2004;87(1):33-40. 14971532
- Shi, W. G., Qu, L., and Wang, J. W. [Study on interventing effect of puerarin on insulin resistance in patients with coronary heart disease]. Zhongguo Zhong Xi Yi Jie He Za Zhi 2002;22(1):21-24. 12585165
- Chen, J., Xu, J., and Li, J. [Effect of puerarin on fibrinolytic activity and lipid peroxide in patients with coronary heart disease]. Zhongguo Zhong.Xi Yi.Jie He Za Zhi 1999;19(11):649-650. 11783153
- Choo, M. K., Park, E. K., Yoon, H. K., and Kim, D. H. Antithrombotic and antiallergic activities of daidzein, a metabolite of puerarin and daidzin produced by human intestinal microflora. Biol Pharm Bull 2002;25(10):1328-1332. 12392089
- Kim, D. H., Jung, E. A., Sohng, I. S., Han, J. A., Kim, T. H., and Han, M. J. Intestinal bacterial metabolism of flavonoids and its relation to some biological activities. Arch Pharm Res 1998;21(1):17-23. 9875509
- Liu, Q., Lu, Z., and Wang, L. Restrictive effect of puerarin on myocardial infarct area in dogs and its possible mechanism. J Tongji Med Univ 2000;20(1):43-45. 12845754
- Luo, Z. R. and Zheng, B. [Effect of Puerarin on platelet activating factors CD63 and CD62P, plasminogen activator inhibitor and C-reactive protein in patients with unstable angia pectoris]. Zhongguo Zhong Xi Yi Jie He Za Zhi 2001;21(1):31-33. 12577374
- Pan, H. P., Yang, J. Z., Li, L. L., Yi, F., Huang, Z. Q., and Huang, K. W. [Experimental study of puerarin injection on the hemorheology in acute blood-stasis model rats]. Zhongguo Zhong Yao Za Zhi 2003;28(12):1178-1180. 15617506
- Yin, Z. Z. and Zeng, G. Y. [Pharmacology of puerarin. V. Effects of puerarin on platelet aggregation and release of 5-HT from platelets]. Zhongguo Yi Xue Ke Xue Yuan Xue Bao 1981;3 Suppl 1:44-47. 6459876
- Yu, Z., Zhang, G., and Zhao, H. [Effects of Puerariae isoflavone on blood viscosity, thrombosis and platelet function]. Zhong Yao Cai 1997;20(9):468-469. 12572427
- Li, S. M., Liu, B., and Chen, H. F. [Effect of puerarin on plasma endothelin, renin activity and angiotensin II in patients with acute myocardial infarction]. Zhongguo Zhong Xi Yi Jie He Za Zhi 1997;17(6):339-341. 9863126
- Wang, L. Y., Zhao, A. P., and Chai, X. S. [Effects of puerarin on cat vascular smooth muscle in vitro]. Zhongguo Yao Li Xue Bao 1994;15(2):180-182. 8010117
- Zhou, Y., Su, X., Cheng, B., Jiang, J., and Chen, H. [Comparative study on pharmacological effects of various species of Pueraria]. Zhongguo Zhong Yao Za Zhi 1995;20(10):619-21, 640. 8679082
- Hsu, H. H., Chang, C. K., Su, H. C., Liu, I. M., and Cheng, J. T. Stimulatory effect of puerarin on alpha1A-adrenoceptor to increase glucose uptake into cultured C2C12 cells of mice. Planta Med 2002;68(11):999-1003. 12451490
- Lee, D. S., Kim, Y. S., Ko, C. N., Cho, K. H., Bae, H. S., Lee, K. S., Kim, J. J., Park, E. K., and Kim, D. H. Fecal metabolic activities of herbal components to bioactive compounds. Arch Pharm Res 2002;25(2):165-169. 12009030
- Qi, B. L. and Qi, B. M. [Effect of the purariae-isofiavones on estrogen level in normal and ovariectomized rats]. Zhongguo Zhong Yao Za Zhi 2002;27(11):850-852. 12776591
- Zhang, Y., Chen, J., Zhang, C., Wu, W., and Liang, X. Analysis of the estrogenic components in kudzu root by bioassay and high performance liquid chromatography. J Steroid Biochem Mol Biol 2005;94(4):375-381. 15857757
- Zheng, G., Zhang, X., Zheng, J., Meng, Q., and Zheng, D. [Estrogen-like effects of puerarin and total isoflavones from Pueraria lobata]. Zhong Yao Cai 2002;25(8):566-568. 12599693
- Guo, X. G., Chen, J. Z., Zhang, X., and Xia, Q. [Effect of puerarin on L-type calcium channel in isolated rat ventricular myocytes]. Zhongguo Zhong Yao Za Zhi 2004;29(3):248-251. 15706853
- Qian, Y., Li, Z., Huang, L., Han, X., Sun, J., Zhou, H., and Liu, Z. Blocking effect of puerarin on calcium channel in isolated guinea pig ventricular myocytes. Chin Med J (Engl) 1999;112(9):787-789. 11717945
- Ye, H. Y., Qiu, F., Zeng, J., Yiao, X. S., and Lai, F. [Effect of daidzein on antiarrhythmia]. Zhongguo Zhong Yao Za Zhi 2003;28(9):853-856. 15015382
- Zhang, G. Q., Hao, X. M., Dai, D. Z., Fu, Y., Zhou, P. A., and Wu, C. H. Puerarin blocks Na+ current in rat ventricular myocytes. Acta Pharmacol Sin 2003;24(12):1212-1216. 14653946
- Overstreet, D. H., Kralic, J. E., Morrow, A. L., Ma, Z. Z., Zhang, Y. W., and Lee, D. Y. NPI-031G (puerarin) reduces anxiogenic effects of alcohol withdrawal or benzodiazepine inverse or 5-HT2C agonists. Pharmacol Biochem Behav 2003;75(3):619-625. 12895679
- Guerra, M. C., Speroni, E., Broccoli, M., Cangini, M., Pasini, P., Minghett, A., Crespi-Perellino, N., Mirasoli, M., Cantelli-Forti, G., and Paolini, M. Comparison between chinese medical herb Pueraria lobata crude extract and its main isoflavone puerarin antioxidant properties and effects on rat liver CYP-catalysed drug metabolism. Life Sci 11-3-2000;67(24):2997-3006. 11133012
- Keung, W. M. and Vallee, B. L. Kudzu root: an ancient Chinese source of modern antidipsotropic agents. Phytochemistry 1998;47(4):499-506. 9461670
- Benlhabib, E., Baker, J. I., Keyler, D. E., and Singh, A. K. Kudzu root extract suppresses voluntary alcohol intake and alcohol withdrawal symptoms in P rats receiving free access to water and alcohol. J Med Food 2004;7(2):168-179. 15298764
- Lin, R. C. and Li, T. K. Effects of isoflavones on alcohol pharmacokinetics and alcohol-drinking behavior in rats. Am J Clin Nutr 1998;68(6 Suppl):1512S-1515S. 9848526
- Xie, C. I., Lin, R. C., Antony, V., Lumeng, L., Li, T. K., Mai, K., Liu, C., Wang, Q. D., Zhao, Z. H., and Wang, G. F. Daidzin, an antioxidant isoflavonoid, decreases blood alcohol levels and shortens sleep time induced by ethanol intoxication. Alcohol Clin Exp Res 1994;18(6):1443-1447. 7695042
- Lu, X. R., Gao, E., Xu, L. Z., Li, H. Z., Kang, B., Chen, W. N., Chen, S. M., and Chai, X. S. Puerarin beta-adrenergic receptor blocking effect. Chin Med J (Engl) 1987;100(1):25-28. 2885157
- Trisomboon, H., Malaivijitnond, S., Suzuki, J., Hamada, Y., Watanabe, G., and Taya, K. Long-term treatment effects of Pueraria mirifica phytoestrogens on parathyroid hormone and calcium levels in aged menopausal cynomolgus monkeys. J Reprod Dev 2004;50(6):639-645. 15647615
- Xue, X. O., Jin, H., Niu, J. Z., and Wang, J. F. [Effects of extracts of root of kudzu vine on mammary gland and uterus development in rats]. Zhongguo Zhong Yao Za Zhi 2003;28(6):560-562. 15015341
- Jeon, G. C., Park, M. S., Yoon, D. Y., Shin, C. H., Sin, H. S., and Um, S. J. Antitumor activity of spinasterol isolated from Pueraria roots. Exp Mol Med 4-30-2005;37(2):111-120. 15886524
- Jin, W. S., Tan, Y. Y., Chen, Y. G., and Wang, Y. [Determination of puerarin, daidzin and daidzein in root of Pueraria lobata of different origin by HPLC]. Zhongguo Zhong Yao Za Zhi 2003;28(1):49-51. 15015267
- Keung, W. M., Lazo, O., Kunze, L., and Vallee, B. L. Potentiation of the bioavailability of daidzin by an extract of Radix puerariae. Proc Natl Acad Sc USA 4-30-1996;93(9):4284-4288. 8633056
- Lee, K. T., Sohn, I. C., Kim, Y. K., Choi, J. H., Choi, J. W., Park, H. J., Itoh, Y., and Miyamoto, K. Tectorigenin, an isoflavone of Pueraria thunbergiana Benth., induces differentiation and apoptosis in human promyelocytic leukemia HL-60 cells. Biol Pharm Bull 2001;24(10):1117-1121. 11642314
- Prasain, J. K., Jones, K., Brissie, N., Moore, R., Wyss, J. M., and Barnes, S. Identification of puerarin and its metabolites in rats by liquid chromatography-tandem mass spectrometry. J Agric Food Chem 6-16-2004;52(12):3708-3712. 15186086
- Prasain, J. K., Jones, K., Kirk, M., Wilson, L., Smith-Johnson, M., Weaver, C., and Barnes, S. Profiling and quantification of isoflavonoids in kudzu dietary supplements by high-performance liquid chromatography and electrospray ionization tandem mass spectrometry. J Agric Food Chem 7-16-2003;51(15):4213-4218. 12848487
- Wang, C. Y., Huang, H. Y., Kuo, K. L., and Hsieh, Y. Z. Analysis of Puerariae radix and its medicinal preparations by capillary electrophoresis. J Chromatogr A 4-3-1998;802(1):225-231. 9588018
- Li, N., Min, Z. D., and Wu, H. M. New oleanene-type triterpene saponins from Pueraria peduncularis. J Asian Nat Prod Res 2002;4(4):253-257. 12450252
- Lukas, S. E., Penetar, D., Berko, J., Vicens, L., Palmer, C., Mallya, G., Macklin, E. A., and Lee, D. Y. An extract of the Chinese herbal root kudzu reduces alcohol drinking by heavy drinkers in a naturalistic setting. Alcohol Clin Exp Res 2005;29(5):756-762. 15897719
- Rezvani, A. H., Overstreet, D. H., Perfumi, M., and Massi, M. Plant derivatives in the treatment of alcohol dependency. Pharmacol Biochem Behav 2003;75(3):593-606. 12895677
- Lee, J. S. Supplementation of Pueraria radix water extract on changes of antioxidant enzymes and lipid profile in ethanol-treated rats. Clin Chim Acta 2004;347(1-2):121-128. 15313149
- Lin, R. C., Guthrie, S., Xie, C. Y., Mai, K., Lee, D. Y., Lumeng, L., and Li, T. K. Isoflavonoid compounds extracted from Pueraria lobata suppress alcohol preference in a pharmacogenetic rat model of alcoholism. Alcohol Clin Exp Res 1996;20(4):659-663. 8800381
- Jang, M. H., Shin, M. C., Kim, Y. J., Chung, J. H., Yim, S. V., Kim, E. H., Kim, Y., and Kim, C. J. Protective effects of puerariaeflos against ethanol-induced apoptosis on human neuroblastoma cell line SK-N-MC. Jpn J Pharmacol 2001;87(4):338-342. 11829154
- Kim, S. and Fung, D. Y. Antibacterial effect of crude water-soluble arrowroot (Puerariae radix) tea extracts on food-borne pathogens in liquid medium. Lett Appl Microbiol 2004;39(4):319-325. 15355532
- Kaufman, P. B., Duke, J. A., Brielmann, H., Boik, J., and Hoyt, J. E. A comparative survey of leguminous plants as sources of the isoflavones, genistein and daidzein: implications for human nutrition and health. J Altern Complement Med 1997;3(1):7-12. 9395689
- Yanagihara, K., Ito, A., Toge, T., and Numoto, M. Antiproliferative effects of isoflavones on human cancer cell lines established from the gastrointestinal tract. Cancer Res 12-1-1993;53(23):5815-5821. 8242641
- Cervellati, R., Renzulli, C., Guerra, M. C., and Speroni, E. Evaluation of antioxidant activity of some natural polyphenolic compounds using the Briggs-Rauscher reaction method. J Agric Food Chem 12-18-2002;50(26):7504-7509. 12475261
- Jiang, R. W., Lau, K. M., Lam, H. M., Yam, W. S., Leung, L. K., Choi, K. L., Waye, M. M., Mak, T. C., Woo, K. S., and Fung, K. P. A comparative study on aqueous root extracts of Pueraria thomsonii and Pueraria lobata by antioxidant assay and HPLC fingerprint analysis. J Ethnopharmacol 1-4-2005;96(1-2):133-138. 15588661
- Jiang, B., Liu, J. H., Bao, Y. M., and An, L. J. Hydrogen peroxide-induced apoptosis in pc12 cells and the protective effect of puerarin. Cell Biol Int 2003;27(12):1025-1031. 14642535
- Xu, X. H. [Effects of puerarin on fatty superoxide in aged mice induced by D-galactose]. Zhongguo Zhong Yao Za Zhi 2003;28(1):66-69. 15015271
- Xu, X. H. and Zhao, T. Q. Effects of puerarin on D-galactose-induced memory deficits in mice. Acta Pharmacol Sin 2002;23(7):587-590. 12100749
- Jin, L. H., Liu, C. F., and Zeng, Y. [Protective effects of puerarin on radiation injury of experimental rats]. Zhong Xi Yi Jie He Xue Bao 2005;3(1):43-45. 15644160
- Jin, M., Qin, J., and Wu, W. [Clinical study on Tianbaokang injection against oxidative injury of vascular endothelial function in ischemic apoplexy]. Zhong Yao Cai 2003;26(2):148-151. 12795227
- Meng, P., Zhou, D., and Hu, X. [Clinical observation of the treatment of infantile viral myocarditis with puerarin]. Zhongguo Zhong Xi Yi Jie He Za Zhi 1999;19(11):647-648. 11783152
- Sang, H. F., Mei, Q. B., Xu, L. X., Wang, Q., Cheng, H., and Xiong, L. Z. Effect of puerarin on neural function and histopathological damages after transient spinal cord ischemia in rabbits. Chin J Traumatol 2004;7(3):143-147. 15294110
- Shi, R. L. and Zhang, J. J. [Protective effect of puerarin on vascular endothelial cell apoptosis induced by chemical hypoxia in vitro]. Yao Xue Xue Bao 2003;38(2):103-107. 12778743
- Zhu, J. H., Wang, X. X., Chen, J. Z., Shang, Y. P., Zhu, J. H., Guo, X. G., and Sun, J. Effects of puerarin on number and activity of endothelial progenitor cells from peripheral blood. Acta Pharmacol Sin 2004;25(8):1045-1051. 15301738
- Kang, R. X. [The intraocular pressure depressive effect of puerarin]. Zhonghua Yan Ke Za Zhi 1993;29(6):336-339. 7912669
- Lu, X. R., Gao, E., Xu, L. Z., Li, H. Z., Kang, B., Chen, W. N., Chen, S. M., and Chai, X. S. [Blocking effect of puerarin on beta-adrenoceptors of isolated organs and the whole animal]. Zhongguo Yao Li Xue Bao 1986;7(6):537-539. 2886005
- Wu, P., Zeng, F., and Ma, H. X. [Study on effect of Puerarin on nitric oxide system in rats' tissue and its mechanism]. Zhongguo Zhong Xi Yi Jie He Za Zhi 2001;21(3):196-198. 12577337
- Zhu, Z. T., Li, H. Q., Lu, Y., Lou, J. S., and Yao, Z. [Inhibitive effect of puerarin on increased NO production by neonatal cardiomyocytes during hypoxia/reoxygenation injury]. Zhongguo Zhong Yao Za Zhi 2001;26(12):856-859. 12776339
- Bennetau-Pelissero, C., Latonnelle, K. G., Lamothe, V., Shinkaruk-Poix, S., and Kaushik, S. J. Screening for oestrogenic activity of plant and food extracts using in vitro trout hepatocyte cultures. Phytochem Anal 2004;15(1):40-45. 14979526
- Boue, S. M., Wiese, T. E., Nehls, S., Burow, M. E., Elliott, S., Carter-Wientjes, C. H., Shih, B. Y., McLachlan, J. A., and Cleveland, T. E. Evaluation of the estrogenic effects of legume extracts containing phytoestrogens. J Agric.Food Chem 4-9-2003;51(8):2193-2199. 12670155
- Kim, O. S., Choi, J. H., Soung, Y. H., Lee, S. H., Lee, J. H., Ha, J. M., Ha, B. J., Heo, M. S., and Lee, S. H. Establishment of in vitro test system for the evaluation of the estrogenic activities of natural products. Arch Pharm Res 2004;27(9):906-911. 15473658
- Wang, X., Wu, J., Chiba, H., Umegaki, K., Yamada, K., and Ishimi, Y. Puerariae radix prevents bone loss in ovariectomized mice. J Bone Miner Metab 2003;21(5):268-275. 12928827
- Benlhabib, E., Baker, J. I., Keyler, D. E., and Singh, A. K. Composition, red blood cell uptake, and serum protein binding of phytoestrogens extracted from commercial kudzu-root and soy preparations. J Med Food 2002;5(3):109-123. 12495583
- Hsu, F. L., Liu, I. M., Kuo, D. H., Chen, W. C., Su, H. C., and Cheng, J. T. Antihyperglycemic effect of puerarin in streptozotocin-induced diabetic rats. J Nat Prod 2003;66(6):788-792. 12828463
- Chen, W. C., Hayakawa, S., Yamamoto, T., Su, H. C., Liu, I. M., and Cheng, J. T. Mediation of beta-endorphin by the isoflavone puerarin to lower plasma glucose in streptozotocin-induced diabetic rats. Planta Med 2004;70(2):113-116. 14994187
- Shen, P., Liu, M. H., Ng, T. Y., Chan, Y. H., and Yong, E. L. Differential effects of isoflavones, from Astragalus membranaceus and Pueraria thomsonii, on the activation of PPARalpha, PPARgamma, and adipocyte differentiation in vitro. J Nutr 2006;136(4):899-905. 16549448
- Dong, L. P. and Wang, T. Y. Effects of puerarin against glutamate excitotoxicity on cultured mouse cerebral cortical neurons. Zhongguo Yao Li Xue Bao 1998;19(4):339-342. 10375780
- Mercer, L. D., Kelly, B. L., Horne, M. K., and Beart, P. M. Dietary polyphenols protect dopamine neurons from oxidative insults and apoptosis: investigations in primary rat mesencephalic cultures. Biochem Pharmacol 1-15-2005;69(2):339-345. 15627486
- Keung, W. M. and Vallee, B. L. Daidzin and its antidipsotropic analogs inhibit serotonin and dopamine metabolism in isolated mitochondria. Proc Natl Acad Sc USA 3-3-1998;95(5):2198-2203. 9482862
- Ma, L., Xiao, P., Guo, B., Wu, J., Liang, F., and Dong, S. [Cerebral protective effects of some compounds isolated from traditional Chinese herbs]. Zhongguo Zhong Yao Za Zhi 1999;24(4):238-inside. 12205950
- Chen, L., Chai, Q., Zhao, A., and Chai, X. [Effect of puerarin on cerebral blood flow in dogs]. Zhongguo Zhong Yao Za Zhi 1995;20(9):560-2, inside. 8679097
- Zhu, X. Y., Su, G. Y., Li, Z. H., Yue, T. L., Yan, X. Z., and Wei, H. L. [The metabolic fate of the effective components of puerariae. III. The metabolism of puerarin (author's transl)]. Yao Xue Xue Bao 1979;14(6):349-355. 517131
- Wang, C. and Song, Z. In vitro monitoring of nanogram levels of puerarin in human urine using flow injection chemiluminescence. Bioorg Med Chem Lett 8-16-2004;14(16):4127-4130. 15261255
- Kim, D. H., Yu, K. U., Bae, E. A., and Han, M. J. Metabolism of puerarin and daidzin by human intestinal bacteria and their relation to in vitro cytotoxicity. Biol Pharm Bull 1998;21(6):628-630. 9657051
Licensed by Natural Standard Copyright © 2010 by Natural Standard Corporation. All Rights Reserved.
back to Plant Profiler back to top
|