|
Plant Profiler
Cranberry (Vaccinium macrocarpon)
|
|
|
Synonyms / Common Names / Related Terms
Alpha-linolenic acid, American cranberry, anthocyanins, Arandano Americano, Arandano trepador, ascorbic acid, Azo-cranberry®, bear berry, benzoic acid, black cranberry, bog cranberry, catechins, cinnamic acids, citric acid, coumaroyl iridoid glycosides, ellagitannins, Ericaceae (family), European cranberry, flavonol glycosides, flavanols, flavonoids, gallotannins, grosse moosebeere, isokarpalo, Kranbeere, Kronsbeere, large cranberry, lectins, low cranberry, malic acid, marsh apple, moosebeere, mossberry, mountain cranberry, n-3 fatty acids, n-6 fatty acids, Oxycoccus hagerupii, Oxycoccus macrocarpus, Oxycoccus microcarpus, Oxycoccus palustris, Oxycoccus quadripetalus, phenolic acids, pikkukarpalo, preisselbeere, proanthocyanidins, quinic acid, ronce d'Amerique, stilbenes, substituted cinnamic acids, sweetened dried cranberries, trailing swamp cranberry, triterpenoids, Tsuru-kokemomo, ursolic acid, Vaccinium edule, Vaccinium erythrocarpum, Vaccinium hageruppi, Vaccinium microcarpum, Vaccinium occycoccus, Vaccinium plaustre, Vaccinium vitis.
Mechanism of Action
Pharmacology:
- Constituents: Six ounces of cranberry juice cocktail (Ocean Spray®), on average, contains 96 calories, 24g carbohydrates, 0.17g crude fiber, 0.02g benzoic acid, 0.28g citric acid, 0.30g malic acid, and 0.45g quinic acid.23 In 94.5g of fresh frozen cranberries, there are an average of 35 calories, 7.8g carbohydrates, 2.1g crude fiber, 0.01g benzoic acid, 0.52g citric acid, 0.48g malic acid, and 0.53g quinic acid.23 Cranberry powder hard gelatin capsules (6.9g in 12 capsules, tested brand manufactured by Murdock Pharmaceuticals) contains 24 calories, 5g carbohydrates, 0.97g crude fiber, 0.001g benzoic acid, 0.46g citric acid, 0.53g malic acid, and 0.50g quinic acid.23 The cranberry juice used in one study contained seven phenolic acids, with 3- and 5-caffeoylquinic acid being the primary components, and 15 flavonol glycosides, with myricetin-3-galactoside and quercetin-3-galactoside being the most prevalent.30 Other constituents of cranberry include coumaroyl iridoid glycosides, flavonoids (anthocyanins, flavonols, ellagitannins, flavonol glycosides, gallotannins, and proanthocyanidins), substituted cinnamic acids, stilbenes, triterpenoids (ursolic acid and its esters), alpha-linolenic acid, n-6 fatty acids, n-3 fatty acids.1,12,34,35,36,37
- Antibacterial effects: Cranberry is traditionally used to treat or prevent urinary tract infections (UTIs) and many reviews have been written regarding the use of cranberry for this purpose.38,39,40,41 It has been studied in vitro for antibacterial or bacteriostatic properties.42 Cranberry juice at 30-35°C did not have viable cells detected for five to eight days.43 Against specific bacteria, cranberry proanthocyanidins and flavonols inhibited the growth of Streptococcus mutans37 and cranberry phenolics showed antibacterial activity against Helicobacter pylori31. However, another study by Monroy-Torres et al. found that cranberry juice did not affect the growth of Escherichia coli, the bacteria responsible for urinary tract infections.44 To simulate an environment more similar to that found in urinary tract infections, some studies have taken urine from subjects who had ingested cranberry products and tested bacterial growth in this medium.45 Although E. coli is the best-studied organism, in vitro examinations of bacterial adherence to urinary epithelial cells have shown that pre-incubation of Proteus, Pseudomonas, and E. coli with cranberry juice cocktail results in decreased adhesion to epithelial cells.46 This effect has been more pronounced in vitro than in vivo, and cranberry juice has not been found to eradicate the most adherent bacteria. Cranberry supplements (Cranactin® tablet, 400mg three times daily for two days) and ascorbic acid (500mg twice daily for two days) each have decreased deposition rates and numbers of adherent E. coli and Enterococcus faecalis in vitro, but not Pseudomonas aeruginosa, Staphylococcus epidermis, or Candida albicans.47 Recently, investigators used a five-fold concentrated preparation of cranberry juice adjusted to a pH of 7.0 and incubated bacterial strains in broth with cranberry and plain controls. At 24 hours, the cranberry-inoculated broth showed no growth of E. coli, Staphylococcusaureus , Pseudomonas, Klebsiella, and Proteus and decreased growth of Enterococcus faecalis or Salmonella.48 Cranberry has not been shown to be bactericidal.
- Anticancer effects: Possible chemopreventive mechanisms of action by cranberry phytochemicals include induction of apoptosis in tumor cells, cell cycle arrest, reduced ornithine decarboxylase activity, decreased expression of matrix metalloproteinases associated with prostate tumor metastasis, and anti-inflammatory activities including inhibition of cyclooxygenases.49,10 Proanthocyanidin oligomers, flavonol and anthocyanin glycosides and triterpenoids are all likely contributors to the observed anticancer properties and may act in an additive fashion to block carcinogenesis.50 Several in vitro and animal studies using CAL-27 oral cancer, DU145 androgen-independent prostate carcinoma, HCT116 colon carcinoma, HepG2 human liver cancer, HT-29 colon carcinoma, KB oral cancer, LNCaP prostate tumor, MCF-7 human breast cancer, and U87 glioblastoma multiforme cell lines have shown that cranberry extracts arrest tumor cell proliferation and induce apoptosis with EC50 values of 11.7-87.4mcM.51,52,35,49,34,53 In one animal study, two extracts slowed the growth of explant tumors, and one of the extracts induced complete regression.51
- Anticoagulant effects: In a few case reports and preliminary studies, ingestion of cranberry increased bleeding and increased INR in patients taking warfarin (Coumadin®).3,4,5,6,7,8,9,25 However, studies in healthy volunteers, patients taking warfarin, and laboratory animals indicate that moderate consumption of cranberry (up to 250mL for up to 10 days) does not affect the anticoagulation effects of warfarin or INR.19,26,20,27,28
- Anti-inflammatory effects: Cranberry has shown some anti-inflammatory activities, including the inhibition of cyclooxygenases.10 In addition, a cranberry fraction potently inhibited pro-inflammatory cytokine and chemokine responses induced by lipopolysaccharides in vitro .11
- Antioxidant effects: In in vitro studies, cranberry and its constituents have shown antioxidant activity 54,15,52,35,49,12,55,21, with the antioxidant activity seen specifically by proanthocyanidins54, quercetin, 3,5,7,3',4'-pentahydroxyflavonol-3-O-beta-D-glucopyranoside, 3,5,7,3',4'-pentahydroxyflavonol-3-O-beta-D-galactopyranoside, and 3,5,7,3',4'-pentahydroxyflavonol-3-O-alpha-l-arabinofuranoside52.
- Antiulcer effects: Cranberry phenolics showed antibacterial activity against Helicobacter pylori in vitro.31
- Antiviral effects: In a series of in vitro studies, Lipson et al. noted that a commercially available cranberry fruit juice drink had a non-specific antiviral effect towards unrelated viral species, including, bacteriophages T2 and T4 and the simian rotavirus SA-11.56 In addition, high molecular weight materials from cranberry dose-dependently inhibited influenza virus A and B from hemagglutinating red blood cells and reduced the viruses' infectivity in vitro.57
- Cholesterol effects: In a study of abdominally obese men, ingestion of 250mL of a low-calorie cranberry juice cocktail daily resulted in a significant increase in plasma HDL-cholesterol concentration (+8.6 ± 14.0% vs. 0mL low-calorie cranberry juice cocktail daily; p<0.01), although this increase was not affected by subsequent increased consumption of cranberry juice cocktail.13 LDL and VLDL levels were not affected. In similar studies, cranberry juice supplementation significantly reduced circulating oxidized LDL concentrations, but did not affect plasma lipoprotein-lipid concentrations.14 These results are supported by in vitro study as well.15
- Cytochrome P450 effects: According to case reports and studies in healthy humans and in vitro, cranberry juice does not inhibit cytochrome CYP2C9, CYP1A2, or CYP3A.19,20,28 However, laboratory studies indicate that cranberry juice may inhibit the function of enteric CYP3A.9
- Genitourinary effects: Cranberries contain a group of compounds, called proanthocyanidins, which are condensed tannins (Gray, 2002; Lowe and Fagelman, 2001; Kuzminski, 1996;Hutchinson, 2005, 740; Howell, 2005 741}. These are thought to be the key factors in inhibiting E. coli adherence. Other potential active constituents include galabiose, prunin, and phlorizin.58 In in vitro study, cranberry powder significantly decreased the mean adherence of Escherichia coli to vaginal epithelial cells (p<0.001) and cranberry proanthocyanidin extract significantly decreased the mean adherence of E. coli to primary cultured bladder epithelial cells (p<0.001) in a linear, dose-dependent fashion.59 In studies using urine from subjects who had ingested cranberry products, the urine inhibited the adhesion of uropathogenic E. coli strains in a manner dependent on the dose of cranberry product ingested.60,61,62,63
- Oral plaque formation prevention activity: Data from in vitro and clinical studies suggests that the ability to reduce Streptococci mutans counts in the saliva is due to the anti-adhesion activity of the cranberry constituent.64,65 In addition, an in vitro study found that cranberry constituents may reduce the capacity of Porphyromonas gingivalis to colonize periodontal sites.18
- Periodontal disease prevention activity: Several mechanisms of action for the benefits of cranberry components in reducing oral diseases, including dental caries and periodontitis, have been suggested.66 Inhibition of acid production, attachment, and biofilm formation by Streptococcus mutan, glucan-binding proteins, extracellular enzymes, carbohydrate production, and bacterial hydrophobicity, are all affected by cranberry components. Two in vitro studies found that cranberry fractions dose-dependently inhibited matrix metalloproteinases and intracellular signaling proteins produced by fibroblasts and macrophages associated with periodontal diseases.16,17
- Urinary acidification properties: With consumption of cranberry juice up to 4L daily, hippuric acid excretion increases, as does urine volume, leading to no changes in urine pH and minimal increases in hippuric acid concentration (to a level that is insufficient for bacteriostasis).24 One study showed only transient decreases in urinary pH in three of four subjects.32 A study in 40 normal subjects found urinary pH to decrease by 0.5, to a minimum of 5.4, with consumption of 250mL of 80% cranberry juice three times daily for 12 days.33 A limitation of this study was that the beverage contained 80% cranberry juice, as opposed to commercially available cocktails that are 25-33% juice. Notably, bacteriostasis has been achieved at pH levels of 5.5.
- UTI prophylaxis properties: For many years, it was believe that cranberry prevented urinary tract infections (UTI) by means of urinary acidification. However, it has now been demonstrated that cranberry juice inhibits bacterial adherence to uroepithelial cells by 75% for 60 of 77 E. coli clinical isolates in vitro when controlling for pH.67,68,69,70,71,72 Uropathogenic E. coli adhere to urinary bladder cells because they have substances that mediate adhesion on the hair-like fimbriae or pili on their surfaces. Cranberry juice contains two compounds that have been found to block E. coli adhesion.73,2 One is fructose (present in most fruit juices). It has been found that fructose in guava, pineapple, mango, grapefruit, blueberry, and cranberry juice inhibit the mannose-sensitive type-1 fimbrial adhesin in yeast aggregation assays. The second inhibitor is a high molecular weight compound called proanthocyanidin, found in cranberry and blueberry juices (genus Ericaceae), which acts on the mannose-resistant P fimbriae or pili expressed by uropathogenic E. coli.74,68,75 In vitro, this inhibition is irreversible.
- Urinary salicylate levels activity: In a study of healthy women who were not taking salicylate drugs, consumption of 250mL of cranberry juice three times daily was associated with an increase (p<0.001) of salicyluric and salicylic acids in urine within one week.29 In addition, two weeks after the start of cranberry juice ingestion, the women showed a small but significant (p<0.05) increase in salicylic acid in plasma.
Pharmacodynamics/Kinetics:
- Human studies have not adequately addressed the precise absorption, bioavailability, volume of distribution, metabolism, elimination half-life, onset of action, time to peak, or duration of activity of cranberry.
- In multiple studies of urinary acidification, urine samples were obtained 2-4 hours after ingestion of cranberry juice, although some trials collected 24-hour urine samples (others do not report the time of collection). One study of plasma antioxidant capacity of cranberry juice determined that plasma phenol levels peaked at one hour, and vitamin C concentrations reached a plateau at two hours.22 Since neither of these are the putative active agents for preventing urinary tract infection, the influence of these findings on dosing is not clear.
- Cranberry alters the pharmacodynamics of warfarin with the potential to increase its effects significantly.25 Cranberry significantly increased the area under the INR-time curve by 30% when administered with warfarin compared with treatment with warfarin alone. Cranberry did not alter S- or R-warfarin pharmacokinetics or plasma protein binding. There was some evidence of VKORC1 (not CYP2C9) genotype-dependent interactions with warfarin.
- Metabolism: According to case reports and studies in healthy humans and in vitro, cranberry juice does not significantly inhibit cytochrome CYP2C9, CYP1A2, or CYP3A.19,20,28,76 However, laboratory studies indicate that cranberry juice may inhibit the function of enteric CYP3A.9
- Excretion: Healthy volunteers who ingested 200mL of cranberry juice excreted six of 12 anthocyanins identified in cranberry in their urine at 24 hours, although the maximum anthocyanin excretion occurred between three and six hours after ingestion.77
Pharmacodynamics/Kinetics:
- Human studies have not adequately addressed the precise absorption, bioavailability, volume of distribution, metabolism, elimination half-life, onset of action, time to peak, or duration of activity of cranberry.
- In multiple studies of urinary acidification, urine samples were obtained 2-4 hours after ingestion of cranberry juice, although some trials collected 24-hour urine samples (others do not report the time of collection). One study of plasma antioxidant capacity of cranberry juice determined that plasma phenol levels peaked at one hour, and vitamin C concentrations reached a plateau at two hours.22 Since neither of these are the putative active agents for preventing urinary tract infection, the influence of these findings on dosing is not clear.
- Cranberry alters the pharmacodynamics of warfarin with the potential to increase its effects significantly.25 Cranberry significantly increased the area under the INR-time curve by 30% when administered with warfarin compared with treatment with warfarin alone. Cranberry did not alter S- or R-warfarin pharmacokinetics or plasma protein binding. There was some evidence of VKORC1 (not CYP2C9) genotype-dependent interactions with warfarin.
- Metabolism: According to case reports and studies in healthy humans and in vitro, cranberry juice does not significantly inhibit cytochrome CYP2C9, CYP1A2, or CYP3A.19,20,28,76 However, laboratory studies indicate that cranberry juice may inhibit the function of enteric CYP3A.9
- Excretion: Healthy volunteers who ingested 200mL of cranberry juice excreted six of 12 anthocyanins identified in cranberry in their urine at 24 hours, although the maximum anthocyanin excretion occurred between three and six hours after ingestion.77
References
- Turner, A., Chen, S. N., Nikolic, D., van Breemen, R., Farnsworth, N. R., and Pauli, G. F. Coumaroyl iridoids and a depside from cranberry (Vaccinium macrocarpon). J Nat Prod 2007;70(2):253-258. 17269823
- Raz, R., Chazan, B., and Dan, M. [Cranberry juice and urinary tract infection]. Harefuah 2004;143(12):891-4, 909. 15666710
- Rindone, J. P. and Murphy, T. W. Warfarin-cranberry juice interaction resulting in profound hypoprothrombinemia and bleeding. Am J Ther 2006;13(3):283-284. 16772772
- Niklasson, A. and Andren, L. [Interaction between Waran and cranberry juice]. Lakartidningen 3-15-2006;103(11):853-854. 16613116
- Suvarna, R., Pirmohamed, M., and Henderson, L. Possible interaction between warfarin and cranberry juice. BMJ 12-20-2003;327(7429):1454. 14684645
- Sylvan, L. and Justice, N. P. Possible interaction between warfarin and cranberry juice. Am Fam Physician 9-15-2005;72(6):1000. 16190498
- Grant, P. Warfarin and cranberry juice: an interaction? J Heart Valve Dis 2004;13(1):25-26. 14765835
- Isele, H. [Fatal bleeding under warfarin plus cranberry juice. Is it due to salicylic acid?]. MMW Fortschr Med 3-11-2004;146(11):13. 15152759
- Uesawa, Y. and Mohri, K. Effects of cranberry juice on nifedipine pharmacokinetics in rats. J Pharm Pharmacol 2006;58(8):1067-1072. 16872553
- Neto, C. C. Cranberry and its phytochemicals: a review of in vitro anticancer studies. J Nutr 2007;137(1 Suppl):186S-193S. 17182824
- Bodet, C., Chandad, F., and Grenier, D. Anti-inflammatory activity of a high-molecular-weight cranberry fraction on macrophages stimulated by lipopolysaccharides from periodontopathogens. J Dent Res 2006;85(3):235-239. 16498070
- Neto, C. C. Cranberry and blueberry: evidence for protective effects against cancer and vascular diseases. Mol Nutr Food Res 2007;51(6):652-664. 17533651
- Ruel, G., Pomerleau, S., Couture, P., Lemieux, S., Lamarche, B., and Couillard, C. Favourable impact of low-calorie cranberry juice consumption on plasma HDL-cholesterol concentrations in men. Br J Nutr 2006;96(2):357-364. 16923231
- Ruel, G., Pomerleau, S., Couture, P., Lamarche, B., and Couillard, C. Changes in plasma antioxidant capacity and oxidized low-density lipoprotein levels in men after short-term cranberry juice consumption. Metabolism 2005;54(7):856-861. 15988692
- Chu, Y. F. and Liu, R. H. Cranberries inhibit LDL oxidation and induce LDL receptor expression in hepatocytes. Life Sci 8-26-2005;77(15):1892-1901. 15982671
- Bodet, C., Chandad, F., and Grenier, D. Inhibition of host extracellular matrix destructive enzyme production and activity by a high-molecular-weight cranberry fraction. J Periodontal Res 2007;42(2):159-168. 17305875
- Bodet, C., Chandad, F., and Grenier, D. Cranberry components inhibit interleukin-6, interleukin-8, and prostaglandin E production by lipopolysaccharide-activated gingival fibroblasts. Eur J Oral Sci 2007;115(1):64-70. 17305718
- Labrecque, J., Bodet, C., Chandad, F., and Grenier, D. Effects of a high-molecular-weight cranberry fraction on growth, biofilm formation and adherence of Porphyromonas gingivalis. J Antimicrob Chemother 2006;58(2):439-443. 16735419
- Lilja, J. J., Backman, J. T., and Neuvonen, P. J. Effects of daily ingestion of cranberry juice on the pharmacokinetics of warfarin, tizanidine, and midazolam--probes of CYP2C9, CYP1A2, and CYP3A4. Clin Pharmacol Ther 2007;81(6):833-839. 17392729
- Pham, D. Q. and Pham, A. Q. Interaction potential between cranberry juice and warfarin. Am J Health Syst.Pharm 3-1-2007;64(5):490-494. 17322161
- Vinson, J. A., Bose, P., Proch, J., Al Kharrat, H., and Samman, N. Cranberries and cranberry products: powerful in vitro, ex vivo, and in vivo sources of antioxidants. J Agric Food Chem 7-23-2008;56(14):5884-5891. 18558697
- Pedersen, C. B., Kyle, J., Jenkinson, A. M., Gardner, P. T., McPhail, D. B., and Duthie, G. G. Effects of blueberry and cranberry juice consumption on the plasma antioxidant capacity of healthy female volunteers. Eur J Clin Nutr 2000;54(5):405-408. 10822287t
- Hughes, B. G. and Lawson, L. D. Nutritional content of cranberry products. Am J Hosp Pharm 1989;46(6):1129. 2750758
- Bodel P, Cotran R, and Kass E. Cranberry juice and the antibacterial action of hippuric acid. J Lab & Clin Med 1959;54(6):881-888.
- Mohammed Abdul, M. I., Jiang, X., Williams, K. M., Day, R. O., Roufogalis, B. D., Liauw, W. S., Xu, H., and McLachlan, A. J. Pharmacodynamic interaction of warfarin with cranberry but not with garlic in healthy subjects. Br J Pharmacol 2008;154(8):1691-1700. 18516070
- Li, Z., Seeram, N. P., Carpenter, C. L., Thames, G., Minutti, C., and Bowerman, S. Cranberry does not affect prothrombin time in male subjects on warfarin. J Am Diet Assoc 2006;106(12):2057-2061. 17126638
- Aston, J. L., Lodolce, A. E., and Shapiro, N. L. Interaction between warfarin and cranberry juice. Pharmacotherapy 2006;26(9):1314-1319. V16945054
- Greenblatt, D. J., von Moltke, L. L., Perloff, E. S., Luo, Y., Harmatz, J. S., and Zinny, M. A. Interaction of flurbiprofen with cranberry juice, grape juice, tea, and fluconazole: in vitro and clinical studies. Clin Pharmacol Ther 2006;79(1):125-133. 16413247
- Duthie, G. G., Kyle, J. A., Jenkinson, A. M., Duthie, S. J., Baxter, G. J., and Paterson, J. R. Increased salicylate concentrations in urine of human volunteers after consumption of cranberry juice. J Agric Food Chem 4-20-2005;53(8):2897-2900. 15826036
- Wilson, T., Singh, A. P., Vorsa, N., Goettl, C. D., Kittleson, K. M., Roe, C. M., Kastello, G. M., and Ragsdale, F. R. Human glycemic response and phenolic content of unsweetened cranberry juice. J Med Food 2008;11(1):46-54. 18361737
- Lin, Y. T., Kwon, Y. I., Labbe, R. G., and Shetty, K. Inhibition of Helicobacter pylori and associated urease by oregano and cranberry phytochemical synergies. Appl Environ Microbiol 2005;71(12):8558-8564. 16332847
- Kahn, H. D., Panariello, V. A., Saeli, J., Sampson, J. R., and Schwartz, E. Effect of cranberry juice on urine. J Am Diet.Assoc 1967;51(3):251-254. 6035629
- Kinney, A. B. and Blount, M. Effect of cranberry juice on urinary pH. Nurs Res 1979;28(5):287-290. 38439
- Seeram, N. P., Adams, L. S., Zhang, Y., Lee, R., Sand, D., Scheuller, H. S., and Heber, D. Blackberry, black raspberry, blueberry, cranberry, red raspberry, and strawberry extracts inhibit growth and stimulate apoptosis of human cancer cells in vitro. J Agric Food Chem 12-13-2006;54(25):9329-9339. 17147415
- Parry, J., Su, L., Moore, J., Cheng, Z., Luther, M., Rao, J. N., Wang, J. Y., and Yu, L. L. Chemical compositions, antioxidant capacities, and antiproliferative activities of selected fruit seed flours. J Agric Food Chem 5-31-2006;54(11):3773-3778. 16719495
- Seeram, N. P., Adams, L. S., Hardy, M. L., and Heber, D. Total cranberry extract versus its phytochemical constituents: antiproliferative and synergistic effects against human tumor cell lines. J Agric Food Chem 5-5-2004;52(9):2512-2517. 15113149
- Duarte, S., Gregoire, S., Singh, A. P., Vorsa, N., Schaich, K., Bowen, W. H., and Koo, H. Inhibitory effects of cranberry polyphenols on formation and acidogenicity of Streptococcus mutans biofilms. FEMS Microbiol Lett 2006;257(1):50-56. 19219097
- Nergard, C. S. and Solhaug, V. [Cranberries for prevention of recurrent urinary tract infections]. Tidsskr.Nor Laegeforen. 2-12-2009;129(4):303-304. 19219097
- Head, K. A. Natural approaches to prevention and treatment of infections of the lower urinary tract. Altern Med Rev 2008;13(3):227-244. 18950249
- Valdevenito S JP. [Recurrent urinary tract infection in women]. Rev Chilena Infectol 2008;25(4):268-276. 18769773
- Jepson, R. G. and Craig, J. C. Cranberries for preventing urinary tract infections. Cochrane.Database.Syst.Rev. 2008;(1):CD001321. 18253990
- Avila-Funes, J. A. [Comments to the manuscript: Does cranberry juice have bacteriostatic activity?]. Rev Invest Clin 2005;57(6):822-824. 16708909
- Walker, M. and Phillips, C. A. The growth of Propionibacterium cyclohexanicum in fruit juices and its survival following elevated temperature treatments. Food Microbiol 2007;24(4):313-318. 17189756
- Monroy-Torres, R. and Macias, A. E. [Does cranberry juice have bacteriostatic activity?]. Rev Invest Clin 2005;57(3):442-446. 16187705
- Tong, H., Heong, S., and Chang, S. Effect of ingesting cranberry juice on bacterial growth in urine. Am J Health Syst Pharm 8-1-2006;63(15):1417-1419. 16849705
- Schmidt, D. R. and Sobota, A. E. An examination of the anti-adherence activity of cranberry juice on urinary and nonurinary bacterial isolates. Microbios 1988;55(224-225):173-181. 3063927
- Habash, M. B., Van der Mei, H. C., Busscher, H. J., and Reid, G. The effect of water, ascorbic acid, and cranberry derived supplementation on human urine and uropathogen adhesion to silicone rubber. Can.J Microbiol. 1999;45(8):691-694. 10528401
- Lee, Y. L., Owens, J., Thrupp, L., and Cesario, T. C. Does cranberry juice have antibacterial activity? JAMA 4-5-2000;283(13):1691. 10755494
- Boivin, D., Blanchette, M., Barrette, S., Moghrabi, A., and Beliveau, R. Inhibition of cancer cell proliferation and suppression of TNF-induced activation of NFkappaB by edible berry juice. Anticancer Res 2007;27(2):937-948. 17465224
- Neto, C. C., Amoroso, J. W., and Liberty, A. M. Anticancer activities of cranberry phytochemicals: an update. Mol Nutr Food Res 2008;52 Suppl 1:S18-S27. 18504707
- Ferguson, P. J., Kurowska, E. M., Freeman, D. J., Chambers, A. F., and Koropatnick, J. In vivo inhibition of growth of human tumor lines by flavonoid fractions from cranberry extract. Nutr Cancer 2006;56(1):86-94. 17176222
- He, X. and Liu, R. H. Cranberry phytochemicals: Isolation, structure elucidation, and their antiproliferative and antioxidant activities. J Agric Food Chem 9-20-2006;54(19):7069-7074. 16968064
- Sun, J. and Hai, Liu R. Cranberry phytochemical extracts induce cell cycle arrest and apoptosis in human MCF-7 breast cancer cells. Cancer Lett 9-8-2006;241(1):124-134. 16377076
- Ariga, T. The antioxidative function, preventive action on disease and utilization of proanthocyanidins. Biofactors 2004;21(1-4):197-201. 15630197
- Apostolidis, E., Kwon, Y. I., and Shetty, K. Potential of cranberry-based herbal synergies for diabetes and hypertension management. Asia Pac J Clin Nutr 2006;15(3):433-441. 16837438
- Lipson, S. M., Sethi, L., Cohen, P., Gordon, R. E., Tan, I. P., Burdowski, A., and Stotzky, G. Antiviral effects on bacteriophages and rotavirus by cranberry juice. Phytomedicine 2007;14(1):23-30. 17140784
- Weiss, E. I., Houri-Haddad, Y., Greenbaum, E., Hochman, N., Ofek, I., and Zakay-Rones, Z. Cranberry juice constituents affect influenza virus adhesion and infectivity. Antiviral Res 2005;66(1):9-12. 15781126
- Turner, A., Chen, S. N., Joike, M. K., Pendland, S. L., Pauli, G. F., and Farnsworth, N. R. Inhibition of uropathogenic Escherichia coli by cranberry juice: a new antiadherence assay. J Agric Food Chem 11-16-2005;53(23):8940-8947. 16277386
- Gupta, K., Chou, M. Y., Howell, A., Wobbe, C., Grady, R., and Stapleton, A. E. Cranberry products inhibit adherence of p-fimbriated Escherichia coli to primary cultured bladder and vaginal epithelial cells. J Urol 2007;177(6):2357-2360. 17509358
- Valentova, K., Stejskal, D., Bednar, P., Vostalova, J., Cihalik, C., Vecerova, R., Koukalova, D., Kolar, M., Reichenbach, R., Sknouril, L., Ulrichova, J., and Simanek, V. Biosafety, antioxidant status, and metabolites in urine after consumption of dried cranberry juice in healthy women: a pilot double-blind placebo-controlled trial. J Agric Food Chem 4-18-2007;55(8):3217-3224. 17381122
- Di Martino, P., Agniel, R., David, K., Templer, C., Gaillard, J. L., Denys, P., and Botto, H. Reduction of Escherichia coli adherence to uroepithelial bladder cells after consumption of cranberry juice: a double-blind randomized placebo-controlled cross-over trial. World J Urol 2006;24(1):21-27. 16397814
- Greenberg, J. A., Newmann, S. J., and Howell, A. B. Consumption of sweetened dried cranberries versus unsweetened raisins for inhibition of uropathogenic Escherichia coli adhesion in human urine: a pilot study. J Altern Complement Med 2005;11(5):875-878. 16296921
- Howell, A. B., Reed, J. D., Krueger, C. G., Winterbottom, R., Cunningham, D. G., and Leahy, M. A-type cranberry proanthocyanidins and uropathogenic bacterial anti-adhesion activity. Phytochemistry 2005;66(18):2281-2291. 16055161
- Weiss, E. I., Kozlovsky, A., Steinberg, D., Lev-Dor, R., Bar Ness, Greenstein R., Feldman, M., Sharon, N., and Ofek, I. A high molecular mass cranberry constituent reduces mutans streptococci level in saliva and inhibits in vitro adhesion to hydroxyapatite. FEMS Microbiol Lett 3-12-2004;232(1):89-92. 15019739
- Yamanaka, A., Kimizuka, R., Kato, T., and Okuda, K. Inhibitory effects of cranberry juice on attachment of oral streptococci and biofilm formation. Oral Microbiol Immunol 2004;19(3):150-154. 15107065
- Bodet, C., Grenier, D., Chandad, F., Ofek, I., Steinberg, D., and Weiss, E. I. Potential oral health benefits of cranberry. Crit Rev.Food Sci Nutr 2008;48(7):672-680. 18663617
- Sobota, A. E. Inhibition of bacterial adherence by cranberry juice: potential use for the treatment of urinary tract infections. J Urol 1984;131(5):1013-1016. 6368872
- Howell, A. B., Vorsa, N., Der, Marderosian A., and Foo, L. Y. Inhibition of the adherence of P-fimbriated Escherichia coli to uroepithelial-cell surfaces by proanthocyanidin extracts from cranberries. N Engl J Med 10-8-1998;339(15):1085-1086. 9767006
- Howell, A. B. and Foxman, B. Cranberry juice and adhesion of antibiotic-resistant uropathogens. JAMA 6-19-2002;287(23):3082-3083. 12069670
- Ahuja, S., Kaack, B., and Roberts, J. Loss of fimbrial adhesion with the addition of Vaccinum macrocarpon to the growth medium of P-fimbriated Escherichia coli. J Urol 1998;159(2):559-562. 9649291
- Allison, D. G., Cronin, M. A., Hawker, J., and Freeman, S. Influence of cranberry juice on attachment of Escherichia coli to glass. J Basic Microbiol 2000;40(1):3-6. 10746194
- Lynch, D. M. Cranberry for prevention of urinary tract infections. Am Fam Physician 12-1-2004;70(11):2175-2177. 15606066
- Zafriri, D., Ofek, I., Adar, R., Pocino, M., and Sharon, N. Inhibitory activity of cranberry juice on adherence of type 1 and type P fimbriated Escherichia coli to eucaryotic cells. Antimicrob Agents Chemother 1989;33(1):92-98. 1674106
- Ofek, I., Goldhar, J., Zafriri, D., Lis, H., Adar, R., and Sharon, N. Anti-Escherichia coli adhesin activity of cranberry and blueberry juices. N Engl J Med 5-30-1991;324(22):1599. 1674106
- Ofek, I., Goldhar, J., and Sharon, N. Anti-Escherichia coli adhesin activity of cranberry and blueberry juices. Adv Exp Med Biol 1996;408:179-183. 8895790
- Grenier, J., Fradette, C., Morelli, G., Merritt, G. J., Vranderick, M., and Ducharme, M. P. Pomelo juice, but not cranberry juice, affects the pharmacokinetics of cyclosporine in humans. Clin Pharmacol Ther 2006;79(3):255-262. 16513449
- Ohnishi, R., Ito, H., Kasajima, N., Kaneda, M., Kariyama, R., Kumon, H., Hatano, T., and Yoshida, T. Urinary excretion of anthocyanins in humans after cranberry juice ingestion. Biosci Biotechnol Biochem 2006;70(7):1681-1687. 16861803
Licensed by Natural Standard Copyright © 2010 by Natural Standard Corporation. All Rights Reserved.
back to Plant Profiler back to top
|