|
Plant Profiler
Cramp bark (Viburnum opulus)
|
|
|
Synonyms / Common Names / Related Terms
American guelder-rose, Caprifoliaceae (family), common guelder-rose, cranberry tree, European cranberry bush, guelder rose, pembina, proanthocyanidins, snowball tree, Viburnum opulus, Viburnum opulus L., Viburnum prunifolium L. , viopudial.
Mechanism of Action
Pharmacology:
- Constituents: Cramp bark contains proanthocyanidins and viopudial.2,1 The squeezed berries of cramp bark contain polysaccharides, including galacturonic acid, galactose, arabinose, mannose, and rhamnose.6 Urosolic acid has been extracted from the flowers.7
- Antimicrobial activity: Viburnum opulus has exhibited disinfectant activity.5
- Antioxidant activity: In an animal study, Viburnum opulus increased the activity of superoxide dismutase, catalase, and gluthatione peroxidase.2 In an in vitro study, Viburnum opulus extract also showed antioxidant activity.3
- Antispasmodic activity: In vitro, viopudial isolated from Viburnum opulus has shown antispasmodic effects on smooth muscle.1
- Astringent activity: In an in vitro study, Viburnum opulus extract showed astringent activity.4
- Gastroduodenoprotective activity: In a study of stressed rats, Viburnum opulus PA prevented gastroduodenal mucosal damage.2
- Hypotensive effects: In vitro, viopudial isolated from Viburnum opulus has shown hypotensive effects.1
- Immunostimulating effects: Some polysaccharides from Viburnum opulus berries enhanced phagocytosis, in particular, the phagocytic index and the secretion of lysosomal enzymes with peritoneal macrophages.6
Pharmacodynamics/Kinetics:
- Insufficient available evidence.
References
- Nicholson, J. A., Darby, T. D., and Jarboe, C. H. Viopudial, a hypotensive and smooth muscle antispasmodic from Viburnum opulus. Proc Soc Exp Biol Med 1972;140(2):457-461. 5037580
- Zayachkivska, O. S., Gzhegotsky, M. R., Terletska, O. I., Lutsyk, D. A., Yaschenko, A. M., and Dzhura, O. R. Influence of Viburnum Opulus proanthocyanidins on stress-induced gastrointestinal mucosal damage. J Physiol Pharmacol 2006;57 Suppl 5:155-167. 17218766
- Ryzhikov, M. A. and Ryzhikova, V. O. [Application of chemiluminescent methods for analysis of the antioxidant activity of herbal extracts]. Vopr Pitan 2006;75(2):22-26. 16729755
- Smirnova, A. S. and Iadrova, V. M. [Comparative study of the astringent effect of liquid extracts from Viburnum opulus L]. Farmatsiia 1968;17(4):42-45. 5759831
- Smetankina, P. P. [DISINFECTANT PROPERTIES OF VIBURNUM OPULUS.]. Vestn Dermatol Venerol 1963;37:75. 14068300
- Ovodova, R. G., Golovchenko, V. V., Popov, S. V., Shashkov, A. S., and Ovodov, IuS. [The isolation, preliminary study of structure and physiological activity of water-soluble polysaccharides from squeezed berries of Snowball tree Viburnum opulus]. Bioorg Khim 2000;26(1):61-67. 10806553
- Plouvier, V. [PRESENCE OF URSOLIC ACID IN THE BLOSSOMS OF VIBURNUM OPULUS L. VAR. STERILE DC (CAPRIFOLIACEAE)]. Ann Pharm Fr 1964;22:313-314. 14197758
Licensed by Natural Standard Copyright © 2010 by Natural Standard Corporation. All Rights Reserved.
back to Plant Profiler back to top
|