|
Plant Profiler
Mistletoe (Viscum album)
|
|
|
Synonyms / Common Names / Related Terms
ABNOBAviscum®, Abnovaviscum Quercus (AQ), all-heal, American mistletoe, Australian mistletoe, avuscumine, bird's lime, birdlime mistletoe, devil's fuge, Drudenfuss, Eurixor®, European mistletoe folia visci, galactoside-specific lectin, golden bough, Helixor®, Herbe de qui (French), Hexenbesen, Iscador® QuFrF, Iscador Qu spezial, Isorel®, Leimmistel, Lektinol®, Lignum crusis (Latin), Loranthaceae (family), Mistlekraut (German), Mistelsenker, Mistletein, mistletoe extract PS76A2, mistletoe lectin (ML), mistletoe of the appletree (Malus), mistletoe of the fir (Abies), mistletoe of the pine (Pinus), mistrel, ML-1, mystyldene, Phoradendron flavescens (Pursh.) Nuttal, Phoradendron leucarpum, Phoradendron macrophyllum, Phoradendron serotinum (Raf.), Phoradendron tomentosum (DC) (American mistletoe), Plenosol®, Stripites Visci, Tallo de muerdago, VaQuFrF, Vischio (Italian), Visci, Visci albi folia, Visci albi fructus, Visci albi herba, Visci albi stipites, viscum, Viscum abietis, Viscum album coloratum (Korean mistletoe), Viscus album quercus frischsaft [Qu FrF], Viscum austriacum, Viscum fraxini-2, viscumin, Vogelmistel, Vysorel®, white mistletoe.
Mechanism of Action
Pharmacology:
- Constituents: Plant proteins with high affinity to specific cellular glycoconjugates: Lectin I (D-galactose-and N-acetyl-D-galactosamine, and sialoglycoproteins specific lectin)37,38,39,40,41, Lectin II and III (N-acetyl-D-galactosamine specific proteins)9. Crude mistletoe extract contains three lectins with different molecular weight and specificity.42 These three lectins agglutinate human erythrocytes and react with immunoglobulins. The mistletoe lectin I is a naturally occurring conjugate of a lectin (B chain) and of an enzyme (A chain).43 The cytotoxicity observed with mistletoe lectin I is caused by inhibiting the protein synthesis on the ribosomal level. Prominent properties of the A chain are mitogenicity and inhibition of the protein synthesis in cell-free system. The B chain activates macrophages and release lymphokines from lymphocytes. The contents and proportions of the isoforms in the mistletoe vary with host tree and harvest time of the year.44,45,45 Mistletoe also contains viscalbCBA, chitin binding protein, and viscotoxins (A1-3; B), which are toxic proteins related to the family of thionins, highly basic cysteine-rich polypeptides. The content and proportions of the isoforms are quite constant in mistletoe from various trees and harvest time of the year.3,30 The green parts of mistletoe contain highly esterified D-galacturonan. However, in mistletoe berries, a complex arabinogalactan is present3, as well as phenylpropanoids: syringin, syringenin-apiosylglucoside46,2, Eleutheroside E2, flavanone glycosides, and alkaloids47.
- Antineoplasm/antiproliferative properties: Extensive literature exists on the use of mistletoe in cancer treatment. In vitro (cell-free and cellular preparations) and animal studies have shown Viscumin (lectin-1 or ML-1) to be cytotoxic to 3T3 cells, lethal to mice, and a potent inhibitor of protein synthesis.17,18,19,20,21,22,23,24,25 The lectins may target the carbohydrates on the cells' surface causing agglutinations.26,27,28,29,30,6,10 Lectins can induce apoptosis in lymphocytes (activity: ML-3>ML-2>ML-1), human peripheral blood lymphocytes, human peripheral blood monocytes, murine thymocytes, human monocytic THP-1 cells, and human K 562 tumor cells, and in animal studies31,32,33,34,35, as well as an increase in the activity of natural killer cells and granulocyte phagocytosis in patients with breast cancer; however, in experimental urothelial carcinogenesis in rats, there is a lack of evidence for a modification or inhibitory effect of the immunomodulatory galactoside-specific mistletoe lectin ML-148. Several mechanisms have been proposed (i.e., ML-1 apoptosis of leukemic B- and T-cell line via caspase-8; ML-2 activates c-Jun N-terminal kinase 1) in apoptotic death of U-937 cells. It has not been established whether apoptosis is a primary or secondary event to inhibition of ribosomal protein synthesis.49,45,50,51,52,53 Purified lectins- ML-I (D-galactose specific), ML-II and III (N-acetyl-D-galactosamine-specific) proteins demonstrated cytotoxicity in killing cells, which was mediated by induction of apoptosis as measured by appearance of hypodiploid nuclei (DNA peak) in flow cytometery. Cytotoxicity may also occur as a result of lectins damaging cell membranes with an influx of Ca2+ and cell shrinkage.5 In tumor patients, regular subcutaneous injections of the optimal dose of mistletoe lectin-1 (1mg/kg of body weight, twice a week) yielded statistically significant increases in certain acute phase reactants (C-reactive protein, haptoglobin, ceruloplasmin, C3-complement, albumin, and immunoglobulin IgM) after four weeks of therapy54, eliciting a humoral immune response. IgG, but not IgM antibodies, were detected against lectins and other components.55 Injection of mistletoe extract to eight cancer patients caused changes in the levels of TNF-α IL-1, IL-6.56 Induction of increased beta-endorphin plasma levels, which correlated with enhanced activities of blood natural killer- and T-cells (correlation between immune and neuroendocrine systems) has been noted with ML-1.57,58
- Rhamnogalacturan: Rhamnogalacturan from mistletoe enhanced cytotoxicity of natural killer cells to K-562 cancer cells.59
- Viscotoxins: Granulocyte phagocytosis was stimulated by viscotoxins.60,61,62,63 Viscotoxins have been shown to result in the generation of reactive oxygen intermediates in human lymphocytes, as well as cell death.45
- Antiviral activity: Five patients with chronic hepatitis C (HCV) received Iscador for one year with a 6-20% decrease in viral load and normalization of liver inflammation (two patients) without side effects.16 Two other patients were also in complete remission of their elevated AST and ALT.
- Cardiovascular effects: In European folk medicine, mistletoe was used for cardiovascular disease treatment. The viscotoxins produced reflex bradycardia, negative inotropic effects and vasoconstriction in cardiac muscle of cats, reduced isometric twitch, and produced contracture and progressive depolarization in rabbit heart preparations, a process reversed by calcium.64,65 Phenylpropanoids were proposed to play a role by inhibiting cAMP phosphodiesterase, although the data is currently inconclusive.46,66,2 A study conducted on cats showed that the effect of mistletoe viscotoxin and phoratoxin on blood circulation resulted in reflex bradycardia, negative inotropic effects on the heart, and in high doses vasoconstriction of vessels in skin and skeletal muscle.65
- Endocrine effects: Aqueous extract of mistletoe (1-10mg/mL) evoked a stepwise (1.1-12.2 fold) stimulation of insulin secretion from clonal pancreatic ß-cells.15 The presence of these insulin-releasing natural products in mistletoe may contribute to the reported antidiabetic property of the plant.
- Immunologic effects: Subcutaneous mistletoe extract (ABNOBAviscum Mali, AM) therapy caused a strong proliferation of peripheral blood mononuclear cells (PBMC) in breast cancer patients. During the observation period, this initial proliferation decreased in all patients.67 TNF-alpha or IL-6 was found in all of monocyte-/macrophage cells. IFN-gamma and IL-4 were released in vitro. Furthermore, a shift to Th1- related cytokines could be observed. It was suggested that these changes might favorably influence the tumor growth. Iscador QuFrF and Iscador Qu Special show immunomodulating qualities in HIV-positive patients, healthy non-smokers and cancer patients. Iscador QuS seems to activate the cytotoxic CD8-positive lymphocytes, important in cancer patients.68Clinical application of extracts of Viscum album (Plenosol®) and Echinacea purpurea (Echinacin) can produce a stimulation of cell-mediated immunity (one therapeutic administration followed by a free interval of one week), or can have a depressive action (daily administrations of higher doses). These observations were confirmed by lymphokine production and assay, 3H-thymidine incorporation and a skin test with recall antigens (Multitest Merieux).69 Mistletoe treatment may express significant reduction in defined lymphocyte subset and hence, a possibility of immunosuppression.1 Iscador for altered various immune indices, including lymphocyte subsets, natural killer cell activity, phagocytic and oxidative activities of PMN leukocytes, eosinophilia, and acute phase marker haptoglobin7,8,36,4,56,14,70, whereas Iscador Pini [IP] mistletoe preparation was not shown to induce eosinophilia in healthy individuals, and this may be related to its content of mistletoe lectins. In contrast, exposure to the viscotoxin-enriched extract IP did not result in specific changes of hematologic parameters. After injections of nontoxic doses of purified mistletoe lectin-1 or its carbohydrate-binding subunit, significant increases in parameters of the cellular host defense system, such as natural killer cytotoxicity or large granular lymphocyte frequency were noted.71 General immunological activity has also been noted by others.72,13,73,74,75,76,77,78
Pharmacodynamics/Kinetics:
- A 2004 Phase I trial studied a twice-weekly, one-hour intravenous infusion, of aviscumine (recombinant mistletoe lectin from Viscum album L.); it was observed to have a half-life (T1/2) of 13 minutes. Plasma levels >2ng/mL (mean IC70 of aviscumine in cell lines) were achieved for only 2.3-7.5 hours per infusion. At dose levels >3200ng/kg the Cmax an area under the concentration time curve (AUC) increased in linear fashion with increasing dosages of the drug.11
- A similar study by the same author studied a once-weekly, 24-hour intravenous infusion of aviscumine. The total half-life (T1/2) was calculated as 2.81 hours. The area under the curve (AUC 0-inf) was 19 h*ng/mL (median) for six evaluable patients at the 5mcg/kg dose.12
References
- Bussing, A., Rosenberger, A., Stumpf, C., and Schietzel, M. [Development of lymphocyte subsets in tumor patients after subcutaneous administration of mistletoe extracts]. Forsch Komplementarmed 1999;6(4):196-204. 10529579
- Wagner, H., Jordan, E., and Feil, B. Studies on the standardization of mistletoe preparations. Oncology 1986;43 Suppl 1:16-22. 3027640
- Jordan, E. and Wagner, H. [Detection and quantitative determination of lectins and viscotoxins in mistletoe preparations]. Arzneimittelforschung 1986;36(3):428-433. 3707660
- Gorter, R. W., van Wely, M., Stoss, M., and Wollina, U. Subcutaneous infiltrates induced by injection of mistletoe extracts (Iscador). Am J Ther 1998;5(3):181-187. 10099057
- Bussing, A., Suzart, K., Bergmann, J., Pfuller, U., Schietzel, M., and Schweizer, K. Induction of apoptosis in human lymphocytes treated with Viscum album L. is mediated by the mistletoe lectins. Cancer Lett 1-19-1996;99(1):59-72. 8564930
- Böcher E, Stumpf C, Büssing A, and et al. [Prospective assessment of toxicity of high dose Viscum album L. infusions in patients with progressive malignomas]. Zeitschrift fur Onkologie 1996;28(4):97-106.
- Chernyshov VP, Omelchenko LI, Heusser P, and et al. Immunomodulatory actions of Viscum album (Iscador) in children with recurrent respiratory disease as a result of the Chernobyl nuclear accident. Complementary Therapies in Medicine 1997;5(3):141-146.
- Chernyshov, V. P., Heusser, P., Omelchenko, L. I., Chernyshova, L. I., Vodyanik, M. A., Vykhovanets, E. V., Galazyuk, L. V., Pochinok, T. V., Gaiday, N. V., Gumenyuk, M. E., Zelinsky, G. M., Schaefermeyer, H., and Schaefermeyer, G. Immunomodulatory and clinical effects of Viscum album (Iscador M and Iscador P) in children with recurrent respiratory infections as a result of the Chernobyl nuclear accident. Am J Ther 2000;7(3):195-203. 11317168
- Doser, C., Doser, M., Hulsen, H., and Mechelke, F. Influence of carbohydrates on the cytotoxicity of an aqueous mistletoe drug and of purified mistletoe lectins tested on human T-leukemia cells. Arzneimittelforschung 1989;39(6):647-651. 2775329
- Stirpe, F., Legg, R. F., Onyon, L. J., Ziska, P., and Franz, H. Inhibition of protein synthesis by a toxic lectin from Viscum album L. (mistletoe). Biochem J 9-15-1980;190(3):843-845. 7470084
- Schoffski, P., Riggert, S., Fumoleau, P., Campone, M., Bolte, O., Marreaud, S., Lacombe, D., Baron, B., Herold, M., Zwierzina, H., Wilhelm-Ogunbiyi, K., Lentzen, H., and Twelves, C. Phase I trial of intravenous aviscumine (rViscumin) in patients with solid tumors: a study of the European Organization for Research and Treatment of Cancer New Drug Development Group. Ann Oncol 2004;15(12):1816-1824. 15550588
- Schoffski, P., Breidenbach, I., Krauter, J., Bolte, O., Stadler, M., Ganser, A., Wilhelm-Ogunbiyi, K., and Lentzen, H. Weekly 24 h infusion of aviscumine (rViscumin): a phase I study in patients with solid tumours. Eur J Cancer 2005;41(10):1431-1438. 15913988
- Lenartz D, Schierholz JM, Menzel J, and et al. Influence of complementary mistletoe lectin therapy in the treatment of malignant glioma . Zeitschrift für Onkologie 1999;31(2):44-46.
- Huber, R., Klein, R., Berg, P. A., Ludtke, R., and Werner, M. Effects of a lectin- and a viscotoxin-rich mistletoe preparation on clinical and hematologic parameters: a placebo-controlled evaluation in healthy subjects. J Altern Complement Med 2002;8(6):857-866. 12614536
- Gray, A. M. and Flatt, P. R. Insulin-secreting activity of the traditional antidiabetic plant Viscum album (mistletoe). J Endocrinol 1999;160(3):409-414. 10076186
- Tusenius, K. J., Spoek, J. M., and Kramers, C. W. Iscador Qu for chronic hepatitis C: an exploratory study. Complement Ther Med 2001;9(1):12-16. 11264964
- Ernst, E. Cancer therapy with mistletoe: hope or hype? J Herb Pharmacother 2002;2(1):3-5. 15277101
- Kuttan, G., Vasudevan, D. M., and Kuttan, R. Isolation and identification of a tumour reducing component from mistletoe extract (Iscador). Cancer Lett 8-30-1988;41(3):307-314. 3409209
- Kuttan, G., Vasudevan, D. M., and Kuttan, R. Presence of a receptor for the active component of Iscador in ascites fluid of tumour bearing mice. Cancer Letters 1989;48(3):223-227. 2605571
- Kuttan, G., Vasudevan, D. M., and Kuttan, R. Effect of a preparation from Viscum album on tumor development in vitro and in mice. J Ethnopharmacol 1990;29(1):35-41. 2345458
- Kuttan, G. and Kuttan, R. Immunological mechanism of action of the tumor reducing peptide from mistletoe extract (NSC 635089) cellular proliferation. Cancer Lett 9-30-1992;66(2):123-130. 1394116
- Kuttan, G. and Kuttan, R. Immunomodulatory activity of a peptide isolated from Viscum album extract (NSC 635 089). Immunol Invest 1992;21(4):285-296. 1398778
- Kuttan, G., Menon, L. G., Antony, S., and Kuttan, R. Anticarcinogenic and antimetastatic activity of Iscador. Anticancer Drugs 1997;8 Suppl 1:S15-S16. 9179361
- Mannel, D. N., Becker, H., Gundt, A., Kist, A., and Franz, H. Induction of tumor necrosis factor expression by a lectin from Viscum album. Cancer Immunol Immunother 1991;33(3):177-182. 2044082
- Valentiner, U., Pfuller, U., Baum, C., and Schumacher, U. The cytotoxic effect of mistletoe lectins I, II and III on sensitive and multidrug resistant human colon cancer cell lines in vitro. Toxicology 2-28-2002;171(2-3):187-199. 11836024
- Buck G. [Viscum album in cancer therapy. 10 years of infusion therapy with Isorel (TM)/ Vysorel (TM)]. Zeitschrift fur Allgemeinmedizin 1996;72(6):359-362.
- Gabius HJ, Hostanska K, Gabius S, and et al. Cytokines as mediators in rationally standardized mistletoe therapy [abstract]. Anticancer Research 1990;10:1473-1474.
- Khwaja TA. Biopharmacological studies of different components of Viscum album (mistletoe) [abstract]. Anticancer Research 1990;10(5B):1374-1375.
- Olsnes, S., Stirpe, F., Sandvig, K., and Pihl, A. Isolation and characterization of viscumin, a toxic lectin from Viscum album L. (mistletoe). J Biol Chem 11-25-1982;257(22):13263-13270. 7142144
- Schaller G, Urech K, Grazi G, and et al. Viscotoxin composition of three European subspecies of Viscum album. Planta Med 1998;64:677-678.
- Hajto T, Hostanska K, Fischer J, and et al. Immunomodulatory effects of Viscum album agglutinin-I on natural immunity. Anti-Cancer Drugs 1997;8(Suppl 1):S43-S46.
- Hostanska, K., Hajto, T., Weber, K., and et al. A natural immunity-activating plant lectin, Viscum album agglutinin-I, induces apoptosis in human lymphocytes, monocytes, monocytic THP-1 cells and murine thymocytes. Nat Immun 1996;15(6):295-311. 9523281
- Antony, S., Kuttan, R., and Kuttan, G. Inhibition of lung metastasis by adoptive immunotherapy using Iscador. Immunol Invest 1999;28(1):1-8. 10073677
- Antony, S., Kuttan, R., and Kuttan, G. Role of natural killer cells in iscador mediated inhibition of metastasis by adoptive immunotherapy. Immunol Invest 2000;29(3):219-231. 10933606
- Hamprecht, K., Handgretinger, R., Voetsch, W., and Anderer, F. A. Mediation of human NK-activity by components in extracts of Viscum album. Int J Immunopharmacol 1987;9(2):199-209. 3583510
- Fischer, S., Scheffler, A., and Kabelitz, D. Stimulation of the specific immune system by mistletoe extracts. Anticancer Drugs 1997;8 Suppl 1:S33-S37. 9179365
- Beuth J, Ko HL, Tunggal L, and et al. [Mistletoe lectin in attendant therapy of mammary carcinoma - First clinical experiences]. Medizinische Welt 1993;44(3):217-220.
- Kannan, S., Gabius, H. J., Chandran, G. J., Pillai, M. R., Nalinakumari, K. R., and Nair, M. K. Expression of galactoside-specific endogenous lectins and their ligands in human oral squamous cell carcinoma. Cancer Lett 9-30-1994;85(1):1-7. 7522955
- Kunze, E., Schulz, H., and Gabius, H. J. Inability of galactoside-specific mistletoe lectin to inhibit N-methyl-N-nitrosourea-induced tumor development in the urinary bladder of rats and to mediate a local cellular immune response after long-term administration. J Cancer Res Clin Oncol 1998;124(2):73-87. 9654190
- Wu, A. M., Chin, L. K., Franz, H., Pfuller, U., and Herp, A. Carbohydrate specificity of the receptor sites of mistletoe toxic lectin-I. Biochim Biophys Acta 9-15-1992;1117(2):232-234. 1525184
- Wu, A. M., Song, S. C., Hwang, P. Y., Wu, J. H., and Pfuller, U. Interaction of mistletoe toxic lectin-I with sialoglycoproteins. Biochem Biophys Res Commun 9-14-1995;214(2):396-402. 7545902
- Franz, H., Ziska, P., and Kindt, A. Isolation and properties of three lectins from mistletoe (Viscum album L.). Biochem J 5-1-1981;195(2):481-484. 7316963
- Franz, H. Mistletoe lectins and their A and B chains. Oncology 1986;43 Suppl 1:23-34. 3543782
- Becker, H. Botany of European mistletoe (Viscum album L.). Oncology 1986;43 Suppl 1:2-7. 3808574
- Bussing, A., Stein, G. M., Wagner, M., Wagner, B., Schaller, G., Pfuller, U., and Schietzel, M. Accidental cell death and generation of reactive oxygen intermediates in human lymphocytes induced by thionins from Viscum album L. Eur J Biochem 1999;262(1):79-87. 10231367
- Deliorman D, Calis I, Ergun F, and et al. The comparative studies of phenylpropanoid glycosides of Viscum album subspecies by HPLC. J Liquid Chromatog Related Technol 1999;22:3101-3114.
- Khwaja, T. A., Dias, C. B., and Pentecost, S. Recent studies on the anticancer activities of mistletoe (Viscum album) and its alkaloids. Oncology 1986;43 Suppl 1:42-50. 3543783
- Kunze, E., Schulz, H., Ahrens, H., and Gabius, H. J. Lack of an antitumoral effect of immunomodulatory galactoside-specific mistletoe lectin on N-methyl-N-nitrosourea-induced urinary bladder carcinogenesis in rats. Exp Toxicol Pathol 1997;49(3-4):167-180. 9314050
- Bantel, H., Engels, I. H., Voelter, W., Schulze-Osthoff, K., and Wesselborg, S. Mistletoe lectin activates caspase-8/FLICE independently of death receptor signaling and enhances anticancer drug-induced apoptosis. Cancer Res 5-1-1999;59(9):2083-2090. 10232592
- Elsasser-Beile, U., Lusebrink, S., Grussenmeyer, T., Wetterauer, U., and Schultze-Seemann, W. Comparison of the effects of various clinically applied mistletoe preparations on peripheral blood leukocytes. Arzneimittelforschung 1998;48(12):1185-1189. 9893935
- Fischer, S., Kabelitz, D., Musielski, H., Scheer, R., and Scheffler, A. Synergism between lectins and vesicles of Viscum album L.- detection by biochemical and immunological methods. J Herb Pharmacother 2002;2(1):7-22. 15277102
- Park, R., Kim, M. S., So, H. S., Jung, B. H., Moon, S. R., Chung, S. Y., Ko, C. B., Kim, B. R., and Chung, H. T. Activation of c-Jun N-terminal kinase 1 (JNK1) in mistletoe lectin II-induced apoptosis of human myeloleukemic U937 cells. Biochem Pharmacol 12-1-2000;60(11):1685-1691. 11077051
- Schumacher, U., Stamouli, A., Adam, E., Peddie, M., and Pfuller, U. Biochemical, histochemical and cell biological investigations on the actions of mistletoe lectins I, II and III with human breast cancer cell lines. Glycoconj J 1995;12(3):250-257. 7496139
- Beuth, J., Gabius, H. J., Steuer, M. K., Geisel, J., Steuer, M., Ko, H. L., and Pulverer, G. [Effect of mistletoe lectin therapy on serum level of defined serum proteins (acute phase proteins) in tumor patients]. Med Klin (Munich) 5-15-1993;88(5):287-290. 7687001
- Stettin, A., Schultze, J. L., Stechemesser, E., and Berg, P. A. Anti-mistletoe lectin antibodies are produced in patients during therapy with an aqueous mistletoe extract derived from Viscum album L. and neutralize lectin-induced cytotoxicity in vitro. Klin Wochenschr 9-14-1990;68(18):896-900. 2124308
- Hajto, T., Hostanska, K., and Gabius, H. J. Modulatory potency of the beta-galactoside-specific lectin from mistletoe extract (Iscador) on the host defense system in vivo in rabbits and patients. Cancer Res 9-1-1989;49(17):4803-4808. 2758413
- Heiny BM. [Adjuvant treatment with standardized mistletoe extract reduces leukopenia and improves the quality of life of patients with advanced carcinoma of the breast getting palliative chemotherapy (VEC regimen)]. Krebsmedizin 1991;12:3-14.
- Heiny, B. M. and Beuth, J. Mistletoe extract standardized for the galactoside-specific lectin (ML-1) induces beta-endorphin release and immunopotentiation in breast cancer patients. Anticancer Res 1994;14(3B):1339-1342. 8067703
- Mueller, E. A. and Anderer, F. A. Chemical specificity of effector cell/tumor cell bridging by a Viscum album rhamnogalacturonan enhancing cytotoxicity of human NK cells. Immunopharmacology 1990;19(1):69-77. 2407685
- Ribereau-Gayon, G., Dumont, S., Muller, C., Jung, M. L., Poindron, P., and Anton, R. Mistletoe lectins I, II and III induce the production of cytokines by cultured human monocytes. Cancer Lett 12-3-1996;109(1-2):33-38. 9020900
- Ribereau-Gayon, G., Jung, M. L., Frantz, M., and Anton, R. Modulation of cytotoxicity and enhancement of cytokine release induced by Viscum album L. extracts or mistletoe lectins. Anticancer Drugs 1997;8 Suppl 1:S3-S8. 9179359
- Stein, G. M., Pfuller, U., and Schietzel, M. Viscotoxin-free aqueous extracts from European mistletoe (Viscum album L.) stimulate activity of human granulocytes. Anticancer Res 1999;19(4B):2925-2928. 10652574
- Stein, G. M., Schaller, G., Pfuller, U., Wagner, M., Wagner, B., Schietzel, M., and Bussing, A. Characterisation of granulocyte stimulation by thionins from European mistletoe and from wheat. Biochim Biophys Acta 1-4-1999;1426(1):80-90. 9878694
- Andersson, K. E. and Johannsson, M. Effects of viscotoxin on rabbit heart and aorta, and on frog skeletal muscle. Eur J Pharmacol 1973;23(3):223-231. 4542875
- Rosell, S. and Samuelsson, G. Effect of mistletoe viscotoxin and phoratoxin on blood circulation. Toxicon 1966;4(2):107-110. 6005043
- Sauvial MP. Effect of phoratoxin B, a toxin isolated from mistletoe, on frog skeletal muscle fibers. Toxicon 1990;28:83-89.
- Stein, G. M. and Berg, P. A. Modulation of cellular and humoral immune responses during exposure of healthy individuals to an aqueous mistletoe extract. Eur J Med Res 6-17-1998;3(6):307-314. 9620894
- Gorter R. [Phase I/II trial to document immunomodulatory, cytotoxic and potentially anti-HIV properties of Iscador]. Merkurstab 1995;2:124-128.
- Coeugniet, E. G. and Elek, E. Immunomodulation with Viscum album and Echinacea purpurea extracts. Onkologie 1987;10(3 Suppl):27-33. 3309759
- Huber, R., Rostock, M., Goedl, R., Ludtke, R., Urech, K., and Klein, R. Immunologic effects of mistletoe lectins: a placebo-controlled study in healthy subjects. J Soc Integr Oncol 2006;4(1):3-7. 16737665
- Hajto, T., Hostanska, K., Frei, K., Rordorf, C., and Gabius, H. J. Increased secretion of tumor necrosis factors alpha, interleukin 1, and interleukin 6 by human mononuclear cells exposed to beta-galactoside-specific lectin from clinically applied mistletoe extract. Cancer Res 6-1-1990;50(11):3322-3326. 2334925
- Lenartz, D., Andermahr, J., Plum, G., Menzel, J., and Beuth, J. Efficiency of treatment with galactoside-specific lectin from mistletoe against rat glioma. Anticancer Res 1998;18(2A):1011-1014. 9615756
- Luther, P., Uhlenbruck, G., Reutgen, H., Samtleben, R., Sehrt, I., and Ribereau-Gayon, G. Are lectins of Viscum album interesting tools in lung diseases? A review of recent results. Z Erkr Atmungsorgane 1986;166(3):247-256. 2428175
- Samal, A. B., Gabius, H. J., and Timoshenko, A. V. Galactose-specific lectin from Viscum album as a mediator of aggregation and priming of human platelets. Anticancer Res 1995;15(2):361-367. 7763007
- Stein, G. and Berg, P. A. Non-lectin component in a fermented extract from Viscum album L. grown on pines induces proliferation of lymphocytes from healthy and allergic individuals in vitro. Eur J Clin Pharmacol 1994;47(1):33-38. 7988621
- Stein, G. M. and Berg, P. A. Evaluation of the stimulatory activity of a fermented mistletoe lectin-1 free mistletoe extract on T-helper cells and monocytes in healthy individuals in vitro. Arzneimittelforschung 1996;46(6):635-639. 8767357
- Stein, G. M., Schietzel, M., and Bussing, A. Mistletoe in immunology and the clinic (short review). Anticancer Res 1998;18(5A):3247-3249. 9858890
- Stein, G. M. and Berg, P. A. Flow cytometric analyses of the specific activation of peripheral blood mononuclear cells from healthy donors after in vitro stimulation with a fermented mistletoe extract and mistletoe lectins. Eur J Cancer 1998;34(7):1105-1110. 9849462
Licensed by Natural Standard Copyright © 2010 by Natural Standard Corporation. All Rights Reserved.
back to Plant Profiler back to top
|