Centricon® Plus-70 Centrifugal Filter
Centricon® Plus-70 centrifugal filterThe Centricon® Plus-70 centrifugal filter is designed for rapid processing of aqueous biological solutions in volumes ranging from 15 to 70 mL. Centricon® Plus-70 filters concentrate most 70 mL solutions down to 350 μL in as little as 25 minutes. Samples are typically concentrated in the 50X to 200X range, depending on the sample type and starting sample volume. These units are a convenient alternative to dialysis, lyophilization, precipitation techniques or stirred cells.
Centrifuge and Rotor Recommendations
 Advantages
  • >90% typical recovery
  • 50X to 200X concentration
  • Low hold-up volume
  • Polypropylene housing minimizes binding
  • True dead stop prevents spinning to dryness
 Applications
  • Concentrating and desalting chromatography column eluates
  • Concentrating monoclonal antibodies
  • Concentrating proteins or viruses from culture supernatants
  • Clarifying tissue homogenates and cell lysates
  • Buffer exchange or diafiltration
Order Centricon® Plus-70 Centrifugal Filters
 Performance
Spin Time with Respect to Filtrate Volume
Spin Time with Respect to Filtrate Volume
 Typical Retenate (Concentrate) Recovery
Membrane NMWL Protein Solute Protein Retentate
10,000 Cytochrome c, 12.4 kDa (0.25 mg/mL) 94
30,000 Bovine Serum Albumin, 67 kDa (1 mg/mL) 96
1,00,000 IgG, 156 kDa (1 mg/mL) 91