Rotor Guide - Amicon® Ultra-15
  • Use any centrifuge and rotor suitable for 50ml tubes.xx
  • Max g-Force 4000 x g for 15 ml in swinging bucket rotor and 5,000 xg for 12 ml in fixed angle rotor
Learn more about Amicon® Ultra-15
 Select Manufacturer to See Recommendations
Note: Centrifuges and rotors listed are not available from Merck. Please confirm product availabilty with the manufacturer.
Beckman
Centrifuge Model Rotor Adapter Required
Allegra X12 FX6100 392268
Allegra X12 SX4750 359475
Allegra X12 SX4750A 359475
back to top
Eppendorf
Centrifuge Model Rotor Adapter Required
5702 A-4-38 22639226
5702 F-34-6-38 226387266
5702 A-44-4 22637665
5702 F-34-6-38 22637266
5702 A-4-62 22638343
back to top
Hermle
Centrifuge Model Rotor Adapter Required
C0200-A C0200-97  
back to top
Hettich
Centrifuge Model Rotor Adapter Required
EBA21 1116 1641
ROTANTA460/460R 5624 5625/4740
ROTANTA460/460R 5624 5620/4840
ROTANTA460/460R 5615 1454
ROTINA35/35R 1724 1430/1384
ROTINA35/35R 1724 1425/1443
ROTINA35/35R 1794 1790/1793
ROTINA35/35R 1717  
ROTINA35/35R 1792 1454
ROTINA35/35R 1720 1454
ROTOFIX32 1624 1381/1384
ROTOFIX32 1619  
ROTOFIX32 1617  
ROTOFIX32 1620A 1454
UNIVERSAL32/32R 1624 1381/1384
UNIVERSAL32/32R 1619  
UNIVERSAL32/32R 1617  
UNIVERSAL32/32R 1620A 1454
back to top
Kendro
Centrifuge Model Rotor Adapter Required
Biofuge Primo/Primo R 75007588 75003103
Biofuge Primo/Primo R 75007555 75007556
Biofuge Stratos 75003057 75003096
Biofuge Stratos 75008172 75008183
Legend Mach 1.6, Multifuge 1/1R 75002006 75003103
Legend Mach 1.6, Multifuge 1/1R 75002002 75007577
Legend Mach 1.6, Multifuge 1/1R 75002001 75005386
Legend T/RT, Multifuge 3/3R 75003057 75003103
Legend T/RT, Multifuge 3/3R 75006441 75006533
Legend T/RT, Multifuge 3/3R 75006446 75006461
back to top
Thermo/Forma
Centrifuge Model Rotor Adapter Required
5528 400ML GP 5852151, 5852152, 5858012, 5852368  
5529 400ML GP 5800236 5802091
5530/5532 1L GP/GPR 5852151, 5852152, 5859581, 5858012, 5858162 (5850968 - for use with rotors listed to left)
5530/5532 1L GP/GPR 5800243 5806560
5677/5678 1L MP/MPR 5852151, 5852152, 5858012, 5858162  
5540/5542 1L MP/MPR 5806555 5806560
5540/5542 1L MP/MPR 5807685 5806651
5694/5696 1L HP/HPR 5808685 5806651
5694/5696 1L HP/HPR 5808850  
5694/5696 1L HP/HPR 5808947 5806560
5660/5662 2500 GP/GPR 5806554 5807005 (sealed), 5805805 (unsealed)
5681/5682 3L 4500 GP/GPR (bench), 5687/5688 3L 4500 GP/GPR (floor) 5800216, 5800228 5805805
5681/5682 3L 4500 GP/GPR (bench), 5687/5688 3L 4500 GP/GPR (floor) 5800218 5805805
5681/5682 3L 4500 GP/GPR (bench), 5687/5688 3L 4500 GP/GPR (floor) 5800825  
back to top
Thermo/IEC
Centrifuge Model Rotor Adapter Required
Clinical 52151, 58012  
Centra® CL2 52151, 52152, 58012, 52368,  
Centra CL3 236 2091S
HN-SII, Centra® HN 215, 958; 52151; 59581 325/320
HN-SII, Centra HN 958; 52152; 59582 326/320
HN-SII, Centra HN 245, 268 7231
HN-SII, Centra HN 58012, 58162  
Centra® CL4 215 325/320
Centra CL4 215 326/320
Centra CL4 224, 52242 7231
Centra CL4 52151, 52152, 58012, 58162  
Centra CL3/CL3R 52151, 52152, 59581, 58012, 58162 (50968 - for use with rotors listed to left)
Centra CL3/CL3R 243 6560E
Centra CL5/CL5R 6555C 6560E
Centra CL5/CL5R 7685C 6651E
Multi/ Multi RF 8685 6651E
Multi/ Multi RF 8850  
Multi/ Multi RF 8947 6560E
Centra® 7/7R 210, 216 7005E (sealed), 5805 (unsealed)
Centra 7/7R 825S  
Centra 8/8R 210, 216 7005E (sealed), 5805 (unsealed)
Centra 8/8R 825S  
Centra GP6/GP6R 6554C 7005E (sealed), 5805 (unsealed)
Centra® GP8/GP8R (Bench),GP8K/GP8RK (Kneewell), GP8F/GP8RF (Floor) 216, 228 5805
Centra GP8/GP8R (Bench),GP8K/GP8RK (Kneewell), GP8F/GP8RF (Floor) 218A 5805
Centra GP8/GP8R (Bench),GP8K/GP8RK (Kneewell), GP8F/GP8RF (Floor) 825S  
Model K, EXD 219A, 256, 949 5805
Model K, EXD 284, 259 384S/7231
Model K, EXD 921, 269, 253 325/305
Model K, EXD 921, 269, 253 325/320
Model K, EXD 921, 269, 253 326/305
Model K, EXD 921, 269, 253 326/320
PR-7000M 219A, 256, 949, 966, 981 5805
PR-7000M 284, 259 384S/7231
PR-7000M 269, 253 325/305
PR-7000M 269, 253 325/320
PR-7000M 269, 253 326/305
PR-7000M 269, 253 326/320
PR-7000M 877, 963 7231
back to top