|
||||||||||||||||||||||
|
Sigma is pleased to announce the launch of its AQUA Peptide Library. Select from a library of over 300 readily available AQUA Peptides for even greater convenience and value. Browse The AQUA Peptide Library Download Sequences and Protein IDs
Search the AQUA Library
Enter your sequence in the box below Ex. VFGSQDLTTV View by Sequence and Product Number
Cannot find your AQUA of interest in the library? Custom AQUA Peptides To meet the specific demands of AQUA experimentation, Sigma has developed a specialized custom peptide offering. Custom AQUA Peptides are synthesized using fully labeled 98 atom% 13C and 98 atom% 15N enriched amino acids (one labeled amino acid per peptide) and are stringently tested to ensure high purity (HPLC), accurate molecular weight (ESI-MS), and specific peptide content. To order Custom AQUA Peptides, call 1-800-325-3010 or visit sigma.com/aquaorder. Protein AQUA References
|
|||||||||||||||||||||
|
|
||||||||||||||||||||||