Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Proteomics > Mass Spectrometry > AQUA™ Peptides > Absolute Quantification of Serum Proteome
Mass Spectrometry
Absolute Quantification of Serum Proteome
Jeffrey J. Porter, Justin Wildsmith, Christopher D. Melm, Mark D. Schuchard, Kevin M. Ray, Dian Er Chen, and Graham B.I. Scott
Download pdf version. (1.96 mb PDF)
Abstract
Introduction
Plasma Facts
Methods
Results
Conclusions
References
Abstract
Plasma proteomics holds great promise for the future of biomarker discovery, as well as in vitro diagnostics. Although plasma is readily accessible for analysis, the study of the plasma proteome is fundamentally limited by its vast dynamic range (10 orders of magnitude). Low-abundance proteins, carrying great diagnostic potential, are often obscured by the presence of high-abundance serum proteins. In order to address this problem, Sigma-Aldrich has developed the ProteoPrep® 20, an antibody-based resin capable of depleting 97–98% of the total protein from plasma. Depletion of these high-abundance proteins allows for visualization of proteins co-eluting with, and masked by, the high-abundance proteins and peptides, using LC-MS methods. Absolute Quantification (Protein-AQUA™) is a targeted quantitative proteomics technique that exhibits robust efficacy and is being increasingly utilized for a wide variety of quantitative proteomics studies. By applying the ProteoPrep 20 for depletion of the most abundant proteins, we have enabled the identification and absolute quantification of numerous lower- abundance proteins from human serum. We believe this may potentially provide a multiplexed targeted approach to the discovery of proteins holding biological or diagnostic significance.
back to top
Introduction
The study of the human plasma proteome is an area of great interest, especially for the pharmaceutical potential of identifying disease biomarkers. Many proteins of pharmaceutical interest appear at low concentrations in the plasma and are, therefore, difficult to detect. Identification of potential biomarkers is especially difficult due to the presence of higher- abundance proteins. Depletion of these high-abundance proteins allows for visualization of proteins co-eluting with, and masked by, the high-abundance proteins and peptides, using LC-MS methods. By removing the high-abundance proteins, higher loads of lower-abundance plasma proteins can then be separated on reverse phase columns, allowing detection and quantification of low copy number proteins. The ProteoPrep 20 Plasma Immunodepletion Kit has been developed for removal of 20 high-abundance proteins from 8 mL of plasma. Depletion of these 20 high-abundance proteins removes greater than 97% of the proteins in plasma and permits analysis of 20- to 50-fold more of each individual protein for improved visualization of lower copy number proteins. By applying the ProteoPrep 20 for depletion of the 20 most abundant proteins, we have enabled the identification and absolute quantification of numerous lower-abundance proteins from human serum.
back to top
Plasma Facts
Plasma Facts
  • The 10 most abundant proteins represent approximately 90% of the total protein mass in human plasma.
  • The 22 most abundant proteins are said to represent approximately 99% of the total protein mass in human plasma.
  • The ProteoPrep 20 plasma immunodepletion column removes the 20 high-abundance plasma/serum proteins listed below. These 20 proteins represent approximately 97% of the total human plasma protein mass.
20 Most Abundant Plasma Proteins
Albumin α-2-Macroglobulin Apolipoprotein A1 Complement C4
IgGs IgMs Apolipoprotein A2 Complement C1 q
Transferrin α-1-Antitrypsin Apolipoprotein B IgDs
IgAs Haptoglobin Ceruloplasmin Plasminogen
Plasma Protein Depletion Workflow
back to top
Methods
High-Abundance Protein Depletion
Twenty high-abundance proteins were depleted from plasma using the ProteoPrep 20 Immunodepletion Kit (PROT20). A large 3.7-mL prototype spin column capable of depleting 100 mL of human plasma was used. The protein concentration of whole and depleted plasma was determined using Bradford assay (B6916), with BSA as the standard.
Tryptic Digestion
Samples of both whole and depleted plasma were prepared in 100 mg aliquots. Each sample was diluted to 0.25 mg/mL in ammonium bicarbonate buffer. Acetonitrile was added to a final concentration of 9% (v/v), and each sample was reduced and alkylated using the ProteoPrep Reduction/Alkylation Kit (PROTRA). Proteomics Grade Trypsin (T6567) was added at a concentration of 1% (w/w) and the sample incubated for 3 hours at 37 °C. An additional aliquot of trypsin at 1% (w/w) was added and then the sample further incubated at 37 °C overnight. The digests were dried to completion in a SpeedVac™.
LC-MS/MS Analysis
An injection of a tryptically digested ProteoPrep 20 depleted plasma sample (prepared as described in Methods) was made using an Agilent capillary 1100 HPLC. The reverse phase separation was performed on a 15 cm × 2.1 mm Discovery C18 column, using a 3-hour gradient. The mobile phase consisted of formic acidified water and acetonitrile. The separation system was coupled to a Thermo Finnigan LTQ linear ion trap mass spectrometer set to perform tandem MS on the ten most abundant ions in each full scan spectrum. After obtaining tandem MS results on an ion of interest, it was added to an exclusion list to facilitate the interrogation of lower-abundance ions. An LC-SRM method was developed corresponding to the protein targets albumin (ideotypic peptide = FQNALLVR) and gelsolin (ideotypic peptide =GASQAGAPQGR), as outlined in the Results section.
AQUA Analysis
An AQUA Peptide stock solution was prepared by dissolving isotopically labeled versions of the two target peptides (FQNALLVR* and GASQAGAPQGR*, respectively) in 0.1% TFA to a final concentration of 62.5 fmol/mL. Each digest sample (whole and depleted plasma) was dissolved in 20 mL of the AQUA Peptide stock solution. Samples were analyzed using an Agilent capillary 1100 HPLC coupled to a Thermo Finnigan LTQ linear ion trap mass spectrometer. Using an LC-SRM method, the absolute quantity of each peptide (and corresponding protein) was determined.
back to top
Results
Overview of the Protein AQUA Method Select an optimal tryptic peptide and stable isotope amino acid from the sequence of your protein of interest.
Order synthetic AQUA Peptide from.
Optimize LC-MS/MS separation protocol for quantitation.
Extract protein from biological samples and add known quantity of AQUA peptide.
Digest.
Analyze by LC-MS/MS or MALDI to quantitate protein of interest.
Figure 1: Overview of the Protein-AQUA Method. This method was developed by Dr. Steve Gygi and colleagues at Harvard Medical School [Stemmann O., Zou H., Gerber S. A., Gygi S. P., Kirschner M. W.; Dual inhibition of sister chromatid separation at metaphase, Cell 2001, Dec 14, 107: 715-726]. Limited use of this method is permitted under a licensing arrangement with Harvard Medical School.
Click here to enlarge
Click on image to enlarge.
Figure 2: Development of a Protein-AQUA LC-SRM for Absolute Protein Quantification. An injection of a tryptically digested ProteoPrep 20 depleted plasma sample (prepared as described in Methods) was made using an Agilent capillary 1100 HPLC coupled to a Thermo Finnigan linear ion trap mass spectrometer. The base peak chromatogram (Figure 2A) shows a complex mixture of species that may or may not have been chromatographically resolved. By harnessing the power of tandem MS, more than 100 proteins were positively identifi ed, with confidence, from the depleted plasma sample. Two of these proteins (albumin and gelsolin) were chosen for further study. The full scan MS shown in Figure 2B illustrates the high abundance of the 481 Da doubly charged ion from one of the albumin peptides identified. This sequence is then confirmed in Figure 2C with an abundance of y- and b-ions. This experimental approach lays the groundwork for choosing an appropriate AQUA peptide in addition to providing the necessary information for designing the SRM experiment. As shown in Figures 2B and 2C, the doubly charged parent ion of this specific albumin peptide was selected for fragmentation and the two most abundant y-ions present in the tandem MS were selected for detection.
back to top
  Protein Identification XC Score Coverage (%)   Protein Identification XC Score Coverage (%)
1 1 Inter-alpha-trypsin inhibitor heavy chain H2 120 21 30 Neuronal kinesin heavy chain 10 16
2 Apolipoprotein A-IV precursor 90 47 31 WIP-related protein 10 24
3 Alpha-1-antichymotrypsin precursor 40 24 32 ATP-binding cassette sub-family F 10 15
4 Inter-alpha-trypsin inhibitor heavy chain H1 40 15 33 NAD(P)H dehydrogenase [quinone] 1 10 6
5 Afamin precursor 40 24 34 Inhibin beta A chain precursor 10 33
6 Kininogen-1 precursor 40 23 35 Corticosteroid-binding globulin precursor 10 2
7 Complement factor H precursor 30 16 36 Neuropeptide FF receptor 1 10 8
8 Serum amyloid P-component precursor 30 35 37 Tripartite motif protein 8 9
9 Histidine-rich glycoprotein precursor 20 15 38 CD180 antigen precursor 8 5
10 Antithrombin-III precursor 20 23 39 CCAAT displacement protein 8 13
11 Gelsolin precursor 20 15 40 Cytoplasmic linker protein 8 15
12 Hemopexin precursor 20 30 41 Eukaryotic translation initiation factor 2 8 25
13 Vitronectin precursor 20 27 42 Sodium channel protein type I alpha subunit 8 5
14 Alpha-2-antiplasmin precursor 20 24 43 Signal peptide peptidase-like 2B 8 9
15 Inter-alpha-trypsin inhibitor heavy chain H4 20 32 44 Acid ceramidase precursor 8 28
16 Angiotensinogen precursor 10 12 45 Glutamate receptor, ionotropic kainate 3 8 16
17 AMBP protein precursor 10 9 46 Gephyrin 8 14
18 Alpha-2-HS-glycoprotein precursor 10 25 47 ARD1 subunit homolog 8 33
19 F-box only protein 31 10 17 48 Low-density lipoprotein receptor protein 2 8 6
20 Prothrombin precursor 10 14 49 Troponin I, fast skeletal muscle 8 15
21 Plasma protease C1 inhibitor precursor 10 8 50 Integrin alpha-E precursor 8 10
22 Tubulin tyrosine ligase-like protein 4 10 7 51 TSC22 domain family protein 1 6 25
23 Heparin cofactor II precursor 10 11 52 Secretogranin-3 precursor 6 12
24 Beta-2-glycoprotein I precursor 10 17 53 BRCA1-associated protein 6 12
25 Isovaleryl-CoA dehydrogenase 10 8 54 Reelin precursor 4 4
26 BLOC-1 subunit 10 14 55 Threonine synthase-like 1 4 19
27 Pantothenate kinase 3 10 14 56 Ciliary dynein heavy chain 5 2 12
28 Contactin 3 precursor 10 8 57 Sorting nexin-19 2 9
29 Complement C5 precursor 10 9        
Table 1: Proteins Identified in LC-MS/MS Analysis of ProteoPrep 20 Depletion Sample (not identified in whole plasma analysis). The proteins listed above were confidently identified in the ProteoPrep 20 depleted sample and were not detected in the whole plasma LC-MS/MS analysis. Using the ProteoPrep 20 depletion technology, we have enabled the identification and absolute quantification of these proteins. Data sets were searched using the SEQUEST algorithm in Bioworks 3.2. Filtration of protein hits consisted of XCORR values >1.9, 2.2, and 3.75 for singly, doubly, and triply charged ions, as well as delta CN > 0.1 and Rsp > 4.
back to top
AQUA-Depletion-Fig-3
Figure 3: Protein-AQUA Analysis of Albumin in Whole Plasma and ProteoPrep 20 Depleted Samples. Selected reaction monitoring (SRM) specifically targets MS detection of the selected parent that produces specific daughter ions. Figures 3A (whole plasma) and 3C (depleted) depict the endogenous albumin peptide, FQNALLVR, as the doubly charged ions fragment to produce two abundant y-ions. These same y-ions are monitored in the AQUA peptide standard shown in Figures 3B (whole plasma) and 3D (depleted); however the absolute mass-to-charge of both parent and daughter ions in the AQUA spike are of higher mass due to the incorporation of a stable isotope amino acid. Quantitation is performed by integrating the co-eluting peak areas shown here at 34.8 minutes. Protein-AQUA analysis of the samples indicates 99.8% depletion of albumin using the ProteoPrep 20 Plasma Immunodepletion Kit, which is in excellent concordance with previously obtained ELISA data.
back to top
AQUA-Depletion-Fig-4
Figure 4: Protein-AQUA Analysis of Gelsolin in Whole Plasma and ProteoPrep 20 Depleted Samples. SRM Analysis of the endogenous gelsolin peptide, GASQAGAPQGR, results in a complex chromatogram in whole plasma (Figure 4A) with poor resolution and low signal-to-noise. When analyzed in depleted plasma (Figure 4C), the endogenous gelsolin peptide is easily separated from interfering species and its concentration readily calculated by comparison to the AQUA peptide internal standard (Figures 4B and 4D).
back to top
Conclusions
Plasma proteomics holds great promise for the future of biomarker discovery, as well as in vitro diagnostics. Although plasma is readily accessible for analysis, the study of the plasma proteome is fundamentally limited by its vast dynamic range (10 orders of magnitude). Low-abundance proteins, carrying great diagnostic potential, are often obscured by the presence of high abundance serum proteins. By applying the ProteoPrep 20 for depletion of the most abundant proteins, we have enabled the identification and absolute quantification of numerous lower-abundance proteins from human serum. We believe this combination of technologies may potentially provide a multiplexed targeted approach to the discovery of proteins holding biological or diagnostic significance.
back to top
References
  1. Anderson, N. L.; Anderson, N. G. The Human Plasma Proteome. Mol. Cell. Proteomics 2002, 1, 845.
  2. Gerber, S. A.; Rush, J.; Stemman, O.; Kirschner, M. W.; Gygi, S. P. Proc. Natl. Acad. Sci. USA 2003, 100, 6940.
back to top
AQUA page
Additional Proteomics & Protein Expression Posters
Proteomics & Protein Expression homepage
Functional Genomics & RNAi homepage