Sigma-Aldrich
Life Science > Proteomics > Mass Spectrometry > AQUA™ Peptides > Protein AQUA Strategy
Mass Spectrometry
Protein AQUA Strategy
In June 2003, Dr. Steve Gygi and his team presented an innovative strategy, Protein AQUA, enabling absolute protein quantitation using stable isotope labeled peptides and HPLC-MS. By applying a common principle, the use of a labeled molecule as an internal standard, to protein analysis, Gygi’s team has advanced the abilities of protein researchers to study complex biological samples quantitatively and has provided a valuable new tool for Proteomics.
An AQUA Peptide is simply a synthetic tryptic peptide corresponding to a peptide of interest. Each AQUA peptide incorporates one stable isotope labeled amino acid, creating a slight increase (6-10 daltons) in molecular weight. When mixed, the native peptide and the synthetic AQUA Peptide elute together chromatographically, migrate together electrophoreticly, and ionize with the same intensity. However, by mass spectrometry, the native peptide and the synthetic AQUA Peptide are easily distinguished.
In a typical AQUA experiment, a known amount of AQUA Peptide is added to a biological protein sample. The sample is then digested and analyzed by HPLC-MS. Extracted ion chromatograms are generated for the native peptide and the synthetic AQUA Peptide internal standard. Using peak ratios, the quantity of native peptide is calculated.
Protein-AQUA is a powerful, enabling technology, the limits of which are only now being explored. For proteomics researchers, it facilitates focused, quantitative studies of not only specific protein expression, but specific amino acid modification as well.
Flow_01
From the Sigma AQUA Peptide Library, select and order an optimal tryptic peptide corresponding to your protein of interest
Flow_03
Optimize your LC-MS/MS protocol to resolve and monitor native and corresponding AQUA peptides
Flow_04
Add a known amount of AQUA Peptide to your biological protein sample
Flow_05
Digest with trypsin
Flow_06
Analyze by LC-MS/MS. Generate extracted ion chromatograms for native and AQUA peptides, and calculate the amount of native peptide present.
back to Protein AQUA page
back to Mass Spectrometry homepage