Life Science > Proteomics > Mass Spectrometry > Proteomics Dynamic Range Standard
Protein Mass Spectrometry
Proteomics Dynamic Range Standard
NEW Proteomics Dynamic Range Standard, UPS2
  • Troubleshoot and optimize your analytical protocol
  • Confirm system suitability before analyzing critical samples
  • Normalize analytical results day-to-day or lab-to-lab
  • Determine your limit of detection
Proteomics Dynamic Range Standard, UPS2
This new standard is an enhancement of Sigma's original Universal Proteomics Standard (UPS1). The same complex mixture of 48 human proteins has been formulated into a dynamic range of concentrations, ranging from 0.5 fmoles to 50 pmoles.
UPS1 | References | 2006 ABRF Study | List of UPS Proteins | UPS1 & UPS2 Home
    Download the new UPS FASTA File (17 Kb)   
Ordering Information
Product No. Product Name Pkg Size Add to Cart
UPS2 Proteomics Dynamic Range Standard 1 set
Shopping Cart
Each set contains one vial of Universal Proteomics Standard and one vial (20 µg) of Proteomics Grade Trypsin (T6567).
The Proteomics Dynamic Range Standard contains the following proteins:
Protein Name [Synonym] MW (Da) (calculated) Amount (fmoles) Protein Name [Synonym] MW (Da) (calculated) Amount (fmoles)
Carbonic anhydrase 1 28,738 50000 Antithrombin-III 49,033 50
Carbonic anhydrase 2 29,095 50000 Beta-2-microglobulin 11,729 50
Complement C5
[Complement C5a]
8,266 50000 BH3 interacting dom1ain death agonist [BID] 21,978 50
Hemoglobin alpha chain 15,127 50000 Cathepsin D 26,624 50
Hemoglobin beta chain 15,867 50000 Glutathione S-transferase A1 [GST A1-1] 25,482 50
Leptin 16,024 50000 Insulin-like growth factor II 7,464 50
Serum albumin 66,393 50000 Platelet-derived growth factor B chain 12,286 50
Ubiquitin 9,387 50000 Thioredoxin 12,424 50
Catalase 59,583 5000 Cathepsin G 26,751 5
Cytochrome b5 16,021 5000 Cytochrome c [Apocytochrome c] 11,608 5
Epidermal growth factor 6,211 5000 Gelsolin 82,954 5
Myoglobin 17,051 5000 Glutathione S-transferase P [GST] 23,220 5
NAD(P)H dehydrogenase [quinone] 1  [DT Diaphorase] 30,984 5000 GTPase HRas [Ras protein] 21,292 5
Peptidyl-prolyl cis-trans isomerase A [Cyclophilin A] 17,947 5000 Serotransferrin [Apotransferrin] 75,143 5
Peroxiredoxin 1 22,106 5000 Ubiquitin-conjugating enzyme E2 C [UbcH10] 20,473 5
Small ubiquitin-related modifier 1 [SUMO-1] 37,420 5000 Ubiquitin-conjugating enzyme E2 E1 [UbcH6] 22,222 5
Alpha-lactalbumin 14,070 500 Annexin A5 35,782 0.5
Creatine kinase M-type
[CK-MM]
43,070 500 C-reactive protein 23,030 0.5
Histidyl-tRNA synthetase [Jo-1] 58,223 500 Fatty acid-binding protein 14,716 0.5
Lysozyme C 14,692 500 Interleukin-8 8,381 0.5
Neddylin [Nedd8] 9,071 500 Lactotransferrin 78,289 0.5
Retinol-binding protein 21,065 500 Microtubule-associated protein tau [Tau protein] 46,810 0.5
Ribosyldihydronicotinamide dehydrogenase (quinone) [Quinone oxidoreductase 2 or NQO2] 25,817 500 Superoxide dismutase
[Cu-Zn]
15,800 0.5
Ubiquitin-conjugating enzyme E2 I [UbcH9] 22,907 500 Tumor necrosis factor
[TNF-alpha]
17,350 0.5
back to top
back to Proteomics & Protein Expression homepage