|
Universal Proteomics Standard, UPS1
- Troubleshoot and optimize your analytical protocol
- Confirm system suitability before analyzing critical samples
- Normalize analytical results day-to-day or lab-to-lab
Developed in collaboration with the ABRF Proteomics Standards Research Group (sPRG), The Universal Proteomics Standard (UPS1) serves as a complex, well-defined, well-characterized reference standard for Proteomics laboratories. As a challenging, complex mixture, the Universal Proteomics Standard contains 48 human proteins (5 pmols of each) ranging in molecular weight from 8,000 to 80,000 daltons. Each constituent protein has been HPLC purified and AAA quantitated prior to formulation.
UPS2 | References | 2006 ABRF Study | List of UPS Proteins | UPS1 & UPS2 Home
| Ordering Information |
| Product No. |
Product Name |
Package Size |
Add to Cart |
| UPS1 |
Universal Proteomics Standard |
1 set |
|
Each set contains one vial of Universal Proteomics Standard and one vial (20 µg) of Proteomics Grade Trypsin (T6567).
The Universal Proteomics Standard contains the following proteins:
| UniProt Accession Number |
UniProt Protein Name [Synonym] |
MW (Da) (calculated) |
Source or recombinant |
Host |
Tag |
Reported PTM's |
| P00709 |
Alpha-lactalbumin |
14,070 |
Milk |
|
|
Glycosylation |
| P08758 |
Annexin A 5 |
35,782 |
Placenta |
|
|
Acetylation |
| P01008 |
Antithrombin-III |
49,033 |
Plasma |
|
|
Glycosylation |
| P61769 |
Beta-2-microglobulin |
11,729 |
Urine |
|
|
|
| P55957 |
BH3 Interacting domain death agonist [BID] |
21,978 |
Recombinant |
E. coli |
|
|
| P00915 |
Carbonic anhydrase 1 |
28,738 |
Erythrocytes |
|
|
Acetylation |
| P00918 |
Carbonic anhydrase 2 |
29,095 |
Erythrocytes |
|
|
Acetylation |
| P04040 |
Catalase |
59,583 |
Erythrocytes |
|
|
|
| P07339 |
Cathepsin D |
26,624 |
Liver |
|
|
Glycosylation |
| P08311 |
Cathepsin G |
26,751 |
Sputum |
|
|
Glycosylation |
| P01031 |
Complement C5 [Complement C5a] |
8,266 |
Recombinant |
E. coli |
|
|
| P02741 |
C-reactive protein |
23,030 |
Plasma |
|
|
|
| P06732 |
Creatine kinase M-type [CK-MM] |
43,070 |
Heart |
|
|
|
| P00167 |
Cytochrome b5 |
16,021 |
Recombinant |
E. coli |
6-His |
|
| P99999 |
Cytochrome c[Apocytochrome c] |
11,608 |
Recombinant |
E. coli |
|
|
| P01133 |
Epidermal Growth Factor |
6,211 |
Recombinant |
E. coli |
|
|
| P05413 |
Fatty acid-binding protein |
14,716 |
Plasma |
|
|
Acetylation, phosphorylation |
| P06396 |
Gelsolin |
82,954 |
Plasma |
|
|
Phosphorylation |
| P08263 |
Glutathione S-transferase A1 [GST A1-1] |
25,482 |
Recombinant |
E. coli |
|
|
| P09211 |
Glutathione S-transferase P [GST] |
23,220 |
Placenta |
|
|
|
| P01112 |
GTPase HRas [Ras protein] |
21,292 |
Recombinant |
E. coli |
|
|
| P69905 |
Hemoglobin alpha chain |
15,127 |
Erythrocytes |
|
|
|
| P68871 |
Hemoglobin beta chain |
15,867 |
Erythrocytes |
|
|
Acetylation, nitrosylation, glycosylation |
| P12081 |
Histidyl-tRNA synthetase [Jo-1] |
58,223 |
Recombinant |
E. coli |
|
|
| P01343 |
Insulin-like growth factor IA |
7,643 |
Recombinant |
E. coli |
|
|
| P01344 |
Insulin-like growth factor II |
7,464 |
Recombinant |
E. coli |
|
|
| P10145 |
Interleukin-8 |
8,381 |
Recombinant |
E. coli |
|
|
| P02788 |
Lactotransferrin |
78,289 |
Milk |
|
|
Glycosylation |
| P41159 |
Leptin |
16,024 |
Recombinant |
E. coli |
|
|
| P61626 |
Lysozyme C |
14,692 |
Milk |
|
|
|
| P10636 |
Microtubule-associated protein tau [Tau protein] |
46,810 |
Recombinant |
E. coli |
6-His |
|
| P02144 |
Myoglobin C |
17,051 |
Heart |
|
|
|
| P15559 |
NAD(P)H dehydrogenase [quinone] 1 [DT Diaphorase] C |
30,984 |
Recombinant |
E. coli |
|
|
| Q15843 |
Neddylin [Nedd8] |
9,071 |
Recombinant |
E. coli |
|
|
| P62937 |
Peptidyl-prolyl cis-trans isomerase A [Cyclophilin A] |
17,947 |
Recombinant |
E. coli |
|
|
| Q06830 |
Peroxiredoxin 1 |
22,106 |
Recombinant |
E. coli |
|
|
| P01127 |
Platelet-derived growth factor B chain |
12,286 |
Recombinant |
E. coli |
|
|
| P02753 |
Retinol-binding protein |
21,065 |
Urine |
|
|
|
| P16083 |
Ribosyldihydronicotinamide dehydrogenase [quinone] [Quinone oxidoreductase 2] [NQO2] |
25,817 |
Recombinant |
E. coli |
|
|
| P02787 |
Serotransferrin [Apotransferrin] |
75,143 |
Plasma |
|
|
Glycosylation |
| P02768 |
Serum Albumin |
66,393 |
Recombinant |
Pichia pastoris |
|
|
| P63165 |
Small ubiquitin-related modifier 1 [SUMO-1] |
37,420 |
Recombinant |
E. coli |
GST |
|
| P00441 |
Superoxide dismutase [Cu-Zn] |
15,800 |
Erythrocytes |
|
|
Acetylation |
| P10599 |
Thioredoxin |
12,424 |
Recombinant |
E. coli |
6-His |
|
| P01375 |
Tumor necrosis factor [TNF-alpha] |
17,350 |
Recombinant |
E. coli |
|
|
| P62988 |
Ubiquitin |
9,387 |
Recombinant |
E. coli |
6-His |
|
| P63279 |
Ubiquitin-conjugating enzyme E2 I [UbcH9] |
22,907 |
Recombinant |
E. coli |
|
|
| O00762 |
Ubiquitin-conjugating enzyme E2 C [UbcH10] |
20,473 |
Recombinant |
E. coli |
6-His |
|
| P51965 |
Ubiquitin-conjugating enzyme E2 E1 [UbcH6] |
22,222 |
Recombinant |
E. coli |
6-His |
|
back to top back to Proteomics & Protein Expression homepage
|