Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Glycosylation
Glycoprofile II
Product Code PP0201
GlycoProfile™ II has been optimized to provide a convenient and reproducible method to remove N-linked glycans from glycoproteins and is compatible with subsequent MALDI-TOF mass spectrometric analysis without interference from any of the reaction components. Reaction conditions may require optimization of denaturation time, reaction time, and PNGase F concentration for deglycosylation of different glycoproteins.
GlycoProfile II contains sufficient enzyme, glycoprotein standard, and reagents for a minimum of 20 reactions when the sample size is between one to two mg of a typical glycoprotein.
Components:
Proteomics Grade PNGase F
Ribonuclease B (RNase B)-glycoprotein standard
10X Reaction Buffer
Octyl b-D-glucopyranoside
2-Mercaptoethanol - 0.90 ml (Product Code M 6250)
pp0201-in-solution
GlycoProfile™ II:
pp0201-maldi-tof
back to Glycosylation