|
Protein Phosphorylation
PhosDecor Kit (PDECOR)
|
Quick & Selective Identification of Phosphorylated Proteins Phosphorylation is an important covalent post-translational modification (PTM) in cell signaling pathways. It is estimated there are more than 100,000 sites of phosphorylation in the human proteome. Phosphorylation of a single protein can alter an entire signal transduction pathway, making each phosphorylation event potentially significant. Traditional identification of phosphorylated protein involves the use of radioisotopes or antibodies, both of which have significant costs and limitations.
- Complete in as little as 4 hours - provides quick identification of protein phosphorylation
- Gels may be dual stained for total protein - measure variations in gel loading and identify non-phosphorylation sites
- Linear detection of phosphoproteins over a wide range - allows for qualitative comparisons
- Compatible with downstream analyses such as in-gel trypsin digestion and MALDI-MS analysis - allows identification of specific phosphorylation sites within the protein
- A complete solution kit - saves time and reduces waste
Sigma now provides a quick and selective identification method for detection of phosphorylated proteins directly in SDS-polyacrylamide gels with the PhosDecor Fluorescent Phosphoprotein In-Gel Detection Kit, (PDECOR). This technology is particularly useful in screening for phosphoproteins in complex protein samples. The kit includes sufficient reagents for staining five mini-gels (8 cm x 8 cm x 1 mm). It contains stain, fixing solution, destain solution, and control.
| Ordering Information |
| Product # |
Product Name |
Pkg Size |
Add to Cart |
| PDECOR |
PhosDecor Fluorescent Phosphoprotein In-Gel Detection Kit |
1 kit |
|
| P7374 |
PhosDecor Stain |
250 ml 1 L |
|
| P7499 |
PhosDecor Control |
1 vial |
|
Procedure | Control Proteins | Linear Detection Over a Wide Range Downstream MALDI-MS Analysis | Related Products
Procedure
 |
PhosDecor™ Fluorescent Phosphoprotein In-Gel Detection Kit Procedure (click on image or above link to view procedure in its entirety.)
|
back to top
Control Proteins
|
The PhosDecor Control contains six proteins, two of which are phosphorylated, providing a set of positive and negative controls for use with PhosDecor Fluorescent Phosphoprotein In-Gel Detection Kit.
| Protein |
MW (kDa) |
Potential Phosphorylation Sites |
| β-galactosidase |
116 |
None |
| Phosphorylase b |
97 |
None |
| Albumin |
66 |
None |
| Ovalbumin |
45 |
2 |
| β-casein |
30 |
5 |
| Lysozyme |
15 |
None |
|
 A: PhosDecor Control stained with PhosDecor Stain B: PhosDecor Control dual stained with EZBlue™ Gel Staining Reagent |
back to top
Linear Detection Over a Wide Range
 Staining of β-casein ranging from 3.13 ng to 200 ng using a UV transilluminator. |
PhosDecor allows for qualitative comparisons in phosphoproteins with comparable phosphorylation states due to a uniform signal response to the presence of phospho groups present. |
back to top
Downstream MALDI-MS Analysis
|
Following PhosDecor staining, protein spots or bands can be digested using the Trypsin Profile IGD Kit (Catalog Number PP0100) and prepared for downstream MALDI-MS analysis. For accurate detection of phosphorylated peptides, digested proteins may need to be enriched prior to mass spectrometry (i.e., PhosphoProfile™ I Phosphopeptide Enrichment Kit, Catalog Number PP0410).
|
 MALDI-MS spectra of ovalbumin stained with PhosDecor Stain. |
back to top
| Related Products |
| Phosphorylation Enrichment |
| Product # |
Product Name |
Add to Cart |
| P9740 |
PHOS-Select™ Iron Affinity Gel |
|
| PP0410 |
Phosphoprofile™ I Phosphopeptide Enrichment Kit |
|
| Total Protein Stains |
| Product # |
Product Name |
Add to Cart |
| G1041 |
EZBlue™ Gel Staining Reagent |
|
| PROTSIL1 |
ProteoSilver™ Silver Stain Kit |
|
| PROTSIL2 |
ProteoSilver™ Silver Stain Plus Kit |
|
| S4942 |
SYPRO® Ruby Protein Gel Stain |
|
| S5692 |
SYPRO® Orange Protein Gel Stain |
|
| In-Gel Trypsin Digestion Kits |
| Product # |
Product Name |
Add to Cart |
| PP0100 |
Trypsin Profile IGD Kit |
|
back to top back to Phosphorylation