Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Proteomics > Protein Chromatography > Affinity Chromatography
Protein Chromatography
Affinity Chromatography
Selective! Image
Affinity chromatography is the preferred method of bioselective adsorption and subsequent recovery of a compound from an immobilized ligand. Each is designed for highly specific and efficient purification of proteins and related compounds. Sigma® offers appropriately selective ligands on beaded and porous matrices for binding target compounds, which are then recovered under mild conditions. Your choice for optimal downstream utility.
Applications for Affinity Media
Activated / Functionalized Matrices
Amino Acid Resins
Avidin Biotin Matrices
Carbohydrate Binding Matrices
Carbohydrate Matrices
Dye Ligand Resins
Glutathione Resins
Heparin Resins
Hydrophobic Interaction Resins
IMAC Matrices
Immunoaffinity Matrices
Nucleotide / Coenzyme Resins
Protein A / Protein G Matrices
Speciality Resins
Online Product Catalog
  Affinity Chromatography Image
back to top
back to Protein Chromatography
Applications for Affinity Media
Affinity Class Potential Application
Activated / Functionalized Functional spacer; support matrix; eliminates handling of toxic reagents
Amino Acid Serum proteins; proteins; peptides; enzymes; rRNA; dsDNA
Avidin Biotin Purification of biotin/avidin & derivatives; biotinylated substances. Biotin derivatives dissociate under nondenaturing conditions.
Carbohydrate Binding Solube glycoproteins ; other carbohydrate-containing substances
Carbohydrate Glycoproteins; lectins; other carbohydrate metabolite proteins. Proper selection can ensure one-step purification.
Dye Ligand Nonspecific interaction. Mimic biological substrates (substrates, cofactors, effetors); proteins. Optimize purification protocol with different dyes
Glutathione Purification of glutathione enzymes and GST tagged recombinant proteins
Heparin General affinity ligand, useful for plasma coagulation proteins, nucleic acid enzymes, lipases, etc.
Hydrophobic Interactions Couple ligands containing free carboxyl groups; proteins
IMAC Immobilized Metal Affinity Chromatography. Uses interactions between protein and chelated metal to separate.
Immunoaffinity Quantitative determinination of antigens, high specificity
Nucleotide / Coenzyme Dehydrogenases; kinases; transaminases. Reliable adsorbents.
Nucleic Acid mRNA; DNA; rRNA; other nucleic acids and oligonucleotides
Protein A / Protein G Purification of immunoglobulins
Speciality Purification of specific classes or types of proteins, coenzymes, or physiological partners
Browse our literature references or submit your own to share!
back to top
Activated / Functionalized Matrices
The availability of pre-activated supports allows the user to quickly and easily custom synthesize affinity resins without having to handle highly reactive reagents. Sigma offers a variety of activated matrices ready for direct coupling to a diversity of ligands. The ligand, generally, must possess a free primary amine, sulfhydryl, or hydroxyl group for direct attachment. Ligand characteristics will be a major determinant in choosing an appropriate matrix.
Historically, cyanogen bromide activated matrices have been very popular. Their ability to efficiently couple amine functionalities under mild conditions has, for many, offset the problems of ligand leaching and ionic properties associated with this chemistry. Sigma offers many alternative activated resins with coupling chemistries suitable for stable attachment to most types of potential ligands. Different activating chemistries will impart unique characteristics to the reactivity and functionality of the affinity matrix. The major properties for each direct activation type are described in the accompanying table.
Direct Activated Matrices
Activation Linkage to Resin Available Reactive Group Specificity of Group Reaction Conditions Bond Type; Stability of Attachment Intrinsic Spacer
Carbonyldimidazole Carbamate Imidazole Carbamate Amine pH 8-10 Carbamate; good stability below pH 10 1 atom neutral
Cyanogen Bromide Final isourea Cyanate ester Amine pH 8-9.5 Isourea; moderately stable 1 atom cationic
Epichlorohydrin Ether Epoxy SH>NH2>OH pH 7-8 SH
pH 9-11 NH2
pH >11 OH
Thioether sec amine ether; all very stable 3 atoms neutral
Epoxy (bis) Ether Epoxy SH>NH2>OH pH 7-8 SH
pH 9-11 NH2
pH >11 OH
Thioether sec amine ether; all very stable 12 atoms neutral
N-Hydroxy-succinimidyl Chloroformate Final carbonate Succinimidyl Carbonate Amine pH 8-9.5 Carbamate; good stability below pH 10 1 atom neutral
p-Nitrophenyl Chloroformate Final carbonate Nitrophenyl Carbonate Amine pH 8.5-10 Carbomate; good stability below pH 10 1 atom neutral
Tresyl Chloride Alkylamine thioether Tresyl sulfonate Amine, thiol pH 7-9.5 Alkylamine thioether; both very stable 0 atoms direct linkage
Vinyl Sulfone Sulfonyl thioether Vinyl sulfone SH>NH2>OH pH 6-8 SH
pH 8-10 NH2
pH >10 OH
ThiotherSec amine ether; Good stability below pH 9 5 atoms neutral, some non-specific effects
Some activated matrices are available which contain an additional spacer due to prior derivatization. These resins may permit the use of milder coupling conditions or a more specific attachment of a ligand. Examples of these are as follows:
Activated Groups Incorporated to Prederivatized Matrices
Active Group Specificity Coupling Conditions Bond to ligand; stability
N-Hydroxysuccinimide Ester; active ester Amine pH 6.0-8.0 Amine; good stability
Disulfide; reactivity based on leaving group Sulfhydryl pH 6.0-8.0 Covalent disulfide; good stability under nonreducing conditions
Sigma offers a variety of matrices with incorporated terminal groups suitable for custom derivatization or coupling. Typically these groups will be a carboxyl or amine function, which may be coupled by an amide bond to a ligand, and require an additional reagent to accomplish condensation. Another group commonly utilized is hydrazide, which may couple to aldehydes by hydrazone formation and typically does not require additional reagent.
References:
  • Hearn, M. T. W. Methods Enzymol. 1987, 135, 102.
  • Kohn, J.; Wilchek, M. Appl. Biochem. Biotechnol. 1984, 9, 285-305.
  • Gilsema, W. J. et al. J. Chrom. 1981, 209, 363-368.
  • Miron, T.; Wilchek, M. Methods Enzymol. 1987, 135, 84-90.
  • Nilsson, K.; Mosbach, K. Methods Enzymol. 1987, 135, 65.
  • Angermann, K.; Barrach, H. J. Anal. Biochem. 1979, 94, 253-258.
  • Lihme, A. et al. J. Chrom. 1986, 376, 299-305.
View our listing of Activated / Functionalized Matrices
back to top
Amino Acid Resins
Sigma offers a variety of amino acids coupled through the amino group to cyanogens bromide activated 4% beaded agarose. These resins may be used directly for adsorpotion of compounds having an affinity for the particular amino acid immobilized, or they can serve as functionalised linkers and allow covalent attachment through their free carboxyl terminus to a ligand containing a free primary amine.
References:
  • Dendriks, D. et al. Biochem. Biophys. Acta 1990, 1034, 86-92.
  • Cleary, S. et al. Biochem. 1989, 28, 188-1891.
View our listing of Amino Acid Resins
back to top
Avidin / Biotin Matrices
The protein Avidin has an extremely high affinity for the cofactor Biotin (Kdiss 10-15M). This characteristic provides a unique and powerful tool, which can be used for separation and purification. Many compounds are easily biotinylated with retention of biological activity, allowing for a broad range of separation applications. Sigma offers a wide variety of biotinylated products, biotinylation reagents and avidin derivates.
One disadvantage of the strong interaction between avidin and biotin is the harsh denaturing conditions required to accomplish dissociation. Harsh conditions may be avoided by using a biotin derivative such as 2-iminobiotin, which dissociates from avidin under milder conditions. Another alternative, is the use of monomeric avidin, which has a much lower affinity for biotin than the natural tetramer. This allows for dissociation of the complex by competitive displacement.
References:
  • Wilchek, M.; Bayer, E. Methods Enzymol. 1990, 184, 5-13 & 194-200.
  • Bayer, E.; Wilchek, M. Meth. Biochem. Anal. 1980, 26, 1.
  • Green, N. M.; Toms, E. J. Biochem. J. 1973, 133, 687.
View our listing of Avidin / Biotin Matrices
back to top
Carbohydrate Binding Matrices
Lectins are the proteins that have the ability to bind to sugar moieties and agglutinate cells. Once immobilized, lectins can be utilized for the purification of selected glycoconjugates, which may generally be recovered by competitive displacement using an inhibitory simple sugar. Lectin resins have been used to:
  • Purify polysaccharides
  • Fractionate cell parts
  • Purify glycoproteins and other glycoconjugate molecules
  • Remove carbohydrate containing impurities from a protein solution
Sugar Specificity Lectin
β-D-gal(1-3)-D-galNAc Arahis hypogaea (Peanut)
Methyl-α-D-gal Artocarpus integrifolia (Jacalin)
α-D-man, α-D-glc Concanavalin A Type VI (Con A)
α-D-man, α-D-glc Succinyl Concanvalin A
Nonreducing end of terminal α-D-mannosyl residue of glycoconjugates Galanthus nivalis (Snowdrop)
D-galNAc Glycine max (Soybean)
D-galNAc Helix pomatia (Roman or edible snail)
α-D-man Lens culinaris (Lentil)
Oligosaccharide Phaseolus vulgaris PHA-E (Red kidney bean)
(D-glcNAc)2, NeuNAc Triticum vulgaris (Wheat germ)
α-L-fuc Ulex europaeus UEA-1 (gorse of Furze)
References:
  • Yamamoto, K. et al. Meth. Mol. Bio. 1993, 14, 17-34.
  • Chilson, O. P.; Kelly-Chilson, A. E. Eur. J. Immunol. 1989, 19, 389-396. (Abstract)
  • Tsuji, T. et al. Mol. Interact. Biosep. 1993, 113-126.
  • Debray, H.; Montrevil, J. Adv. Lectin Res. 1991, 4, 51-96.
View our listing of Carbohydrate Binding Matrices
back to top
Carbohydrate Matrices
Immobilized sugars and sugar derivatives offer an effective means of purification for a variety of proteins, which recognize and bind carbohydrate moieties. Among the most widely used applications for these matrices are the purification of lectins, glycosidases, and carbohydrate directed antibodies. Lectin isolation is of particular interest in that they are highly useful for studies of cell membrane structure and function, determination of blood groups, cell fractionation, and stimulation of lymphocyte mitosis.
References:
  • Vretblad, P. Biochem. Biophs. Acta 1976, 434, 169-176.
  • Matsumoto, I. et al. Anal. Biochem. 1981, 116, 103.
  • Dey, P. M. Eur. J. Biochem. 1984, 140, 385. (Abstract)
View our listing of Carbohydrate Matrices
back to top
Dye Ligand Resins
Dye ligand chromatography is affinity chromatography that utilizes covalently bond textile dye (reactive dyes) to purify proteins. These dyes resemble natural substrates which proteins have affinities for, thus dye ligand chromatography is sometimes referred to as pseudo-affinity chromatography. Immobilized dyes are not specific adsorbents, but rather have a broad binding range for a variety of proteins. After careful investigation of the proper binding and elution conditions, a high degree of purification of a target protein can be attained even in the presence of a mixture of other proteins.
References:
  • Scopes, R. K. Anal. Biochem. 1987, 165, 235-246.
  • Scopes, R. K. J. Chrom. Biomed. Appl. 1986, 376, 131-140. (Abstract)
  • Lowe, C. R.; Pearson, J. C. Methods Enzymol. 1984, 104, 97-112.
  • Stellwagon, E. Methods Enzymol. 1990, 182, 343-357.
  • Miribel, L. et al. J. Biochem. Biophys. Meth. 1988 16, 1-15. (Abstract)
View our listing of Dye Ligand Resins
back to top
Glutathione Resins
Glutathione is a tri-peptide, which consists of the amino acids Glutamic acid, Cysteine and Glycine. Glutathione and its derivatives are very useful as ligands in affinity chromatography for the isolation of Glutathione requiring enzymes. These include detoxification enzymes such as: Glutathione Transferase, Glutathione Peroxidase and Glyoxalase I. In recent years, Glutathione agarose resins have become useful for the isolation of fusion proteins due to the presence of Glutathione transferase (GST) as part of the fusion protein. By fusing GST to recombinant proteins, Glutathione resins can separate a selected target protein from other components.
References:
  • Smith, D. B.; Johnson, S. K. Gene 1988, 67, 31. (Abstract)
  • Guan, K.; Dixon, J. E. Anal. Biochem. 1991, 192, 262. (Abstract)
  • Nabell, L. M. et al. Cell Growth Differ. 1994, 5, 87-93. (Abstract)
  • Irzyk, G. P.; Fuerst, E. P. Plant Physiol. 1993, 102, 803-810. (Abstract)
View our listing of Glutathione Resins
View our listing of Fusion Tags
back to top
Heparin Resins
Heparin is a highly sulphated glycosaminoglycan, which has widespread use as a general affinity ligand. Its high degree of sulfation imparts a strong acidic nature to the molecule, therefore it may bind to many substances by ionic interaction. In addition, heparin contains unique carbohydrate sequences, which act as specific binding sites for some proteins. Immobilized heparin has been used to purify plasma coagulation proteins, nucleic acid enzymes, lipases, and other proteins.
References:
  • Josic, D.; Bal F.; Schwinn, H. J. Chrom. 1993, 632, 1-10. (Abstract)
  • Farooqui, A. A. J. Chrom. Rev. 1980, 184, 335-345.
  • Sasaki, H. et al. J. Chrom. 1987, 400, 123. (Abstract)
  • Mitra, G. et al. Biotechnol. Bioeng. 1986, 28, 217.
View our listing of Heparin Resins
back to top
Hydrophobic Interaction Resins
Hydrophobic chromatography is a versatile technique used for the separation and purification of proteins based on their hydrophobicity. Columns are usually run under conditions that favor hydrophobic interaction, such as high ionic strength. This makes hydrophobic chromatography an ideal tool to use following salt precipitation. Our hydrophobic gels are available in three functional variations:
  • Pure hydrophobic resins—these gels contain ligands attached through a stable ether linkage and they contain no charged regions
  • Mixed property resins—these gels contain ligands attached through an amine group and in some cases they will carry a charge
  • Aminoalkyl resins—these gels provide a hydrophobic spacer to which other functional groups may be coupled
References:
  • Walsh, M. P. et al. Biochem. J. 1984, 224, 117-127. (Abstract)
  • Shartiel, S. Methods Enzymol. 1984, 104, 69.
  • Kennedy, R. M. Methods Enzymol. 1990, 182, 339-343.
View our listing of Hydrophobic Interaction Resins
back to top
Immunoaffinity Matrices
Immobilized antibodies are used as analytical tools for the quantitative determination of antigens, including immunoglobulins and haptens. Immunoprecipitation followed by SDS-PAGE is a technique typically used to determine the quantity and presence of an antigen in a complex protein mixture such as a cell lysate. Cells may be separated according to surface antigens using various immobilized immunochemicals. Antibody-agarose products are also used in a variety of application including immunoadsorption, affinity chromatography and as solid-phase secondary antibodies. Because of this diversity, Sigma offers a wide selection of antibodies and whole sera immobilized onto 4% crosslinked agarose.
References:
  • Rönnstrand, L. et al. J. Biol. Chem. 1987, 262, 2929. (Abstract)
  • Ho, K. J. Biotechnol. Appl. Biochem. 1991, 14, 296-305.
  • Bazin, H.; Malache, J. M. J. Immunol. Meth. 1986, 3, 19-24. (Abstract)
View our listing of Immunoaffinity Matrices
back to top
IMAC Matrices
Immobilized Metal Affinity Chromatography (IMAC) is the process of protein separation based on the differential interaction of various proteins with different chelated / insolubilized metals. This interaction depends on:
  • The relative amount and location of certain amino acids within the protein (namely histidine, cysteine, tyrosine, and typtophan)
  • The type of metal utilized
Generally, different transition metals may be chelated to and exchanged within the same resin, however, some chelating groups may demonstrate preferential chelation of certain metals. Commonly utilized metals are: Copper, Zinc, Iron, and Nickel.
Metal Application
Cobalt Purification of HIS-tagged proteins
Nickel Purification of HIS-tagged proteins
Copper Usually for purification of HIS-tagged proteins and histidine containing peptides
Zinc Usually for the purification of HIS-tagged proteins
Iron Purification of Phosphopeptides
Gallium Purification of Phosphopeptides
References:
  • Porath, J.; Olin, B. Biochem. 1983, 22, 1621. (Abstract)
  • Hutchens, T. W.; Yip, T. T. Anal. Biochem. 1990, 191, 160-168. (Abstract)
  • Yip, T. T.; Hutchens, T. W. Mol. Biotechnol. 1994, 1, 151-164. (Abstract)
View our listing of IMAC Matrices
View our listing of HIS-Select
View our listing of Phos-Select
back to top
Nucleotide / Coenzyme Resins
Cofactors, coenzymes, and substrates bound to matrices play a major role in protein purification. Since nearly one third of known enzymes require a nucleotide coenzyme, nucleotide bound resins are useful for the purification of many proteins. Protein binding to a nucleotide coenzyme is dependent on spacer arm length and the position of ligand attachment. Sigma offers a wide variety of nucleotide resins with different attachment positions and spacer arms o provide the chemist greater flexibility in affinity fractionations.
References:
  • Kiessling, P. et al. Biol. Chem. Hoppe-Seyler 1993, 374, 183-192. (Abstract)
  • Yamazaki, Y. et al. Biochem. 1991, 30, 1503-1509. (Abstract)
  • Ramandham, S. et al. Biochem. 1994, 33, 1442-1452. (Abstract)
  • Gonsky, R. et al. J. Biol. Chem. 1990, 265, 9083-9089. (Abstract)
View our listing of Nucleotide / Coenzyme Resins
back to top
Protein A / Protein G Matrices
Protein A and Protein G are bacterial proteins, which demonstrate specific binding to the Fc (non-antigen binding) portion of many classes of immunoglobulins. Protein A and G affinity matrices have been used primarily for:
  • Affinity purification of immunoglobulin, primarily IgGs
  • Separation of Fc from Fab fragments
Protein A resins have historically been popular for most potential applications, however it has been demonstrated that Protein G resin can enhance and broaden the scope of application. The binding characteristics of the two proteins for various types of immunoglobulins vary and may be used to good advantage. Some of the major differences in binding are:
  • Protein A
    • Broad species reactivity; binds well to IgG from human, rabbit, cow and guinea pig
    • Weak binding to monoclonal antibodies
  • Protein G
    • Broader species reactivity; much stronger binding to IgG from mouse, rat and goat
    • Stronger binding to monoclonal antibodies
References:
  • Akerström, B. et al. J. Immunol. 1985 135, 2589-2592. (Abstract)
  • Coleman, P. L. et al. J. Chrom. 1990, 512, 345-363.
  • Lindmark, R. et al. J. Immunol. Meth. 1983, 62, 1-13. (Abstract)
View our listing of Protein A / Protein G Matrices
back to top
Specialty Resins
The following is a sample list of products containing a variety of specialty affinity matrices. Each resin is listed with possible applications and references.
Resin Type For the purification of (Applications), [Reference]
Albumin agarose Steroids, amino acids, bilirubin & heme peptides (For general hydrophobic adsorption & chiral resolution) [1,2]
p-Aminobenzamide agarose Serine proteases, urokinases, plasminogen activators and protease removal [3,4]
m-Aminophenylboronic acid agarose Glycoproteins (This matrix offers high capacity) [5]
N(p-Aminophenyl)oxamic acid agarose Neuraminidase & influenza virus [6]
Antithrombin III agarose Heparin & thrombin [7,8]
Aprotinin agarose Serine proteases, urokinases & protease removal [9]
Cholesterol hemisuccinate agarose HDL, LDL & cholesterol oxidase [10]
α-Cobrotoxin agarose Nicotinic acetylcholine (snake toxin) receptors [12]
Fetuin agarose Lectins, glycosidases & carbohydrate binding proteins [12,13]
Folic acid agarose Folate binding proteins [14]
Forskolin agarose Adenylate cyclase [15]
Gelatin agarose Fibronectin [16,17]
Hemoglobin agarose Hemoglobin based O2 carriers [18]
L-Histidyldiazobenzyl phophonic acid agarose Phosphatases [19]
Histone agarose Protein kinases [20]
Methotrexate agarose Dihydrofolate reductase [21]
Pepstatin A agarose Cathepsin D & Rennin [22]
Phosphodiesterase 3’-5’ cyclic nucleotide activator agarose Calmodulin binding proteins [23,24]
O-Phospho-L-Tyrosine agarose Phosphatases [19]
Poly-L-Lysine agarose DNA, RNA & T4 phage [25]
Polymyxin B agarose Lipopolysaccharides (endotoxins) [26,27]
Tartaric acid agarose Prostatic acid phosphatases [30,31]
Trypsin inhibitor agarose Proteases or protease removal [32,33]
References:
  1. Popov, D. et al. J. Mol. Cell. Cardiol. 1992, 24, 989-1002.
  2. Allenmark, S.; Bomgren, B.; Boren, H. J. Chrom. 1982, 237, 473.
  3. Winkler, M. E., et al. Biotechnology 1985, 3, 990-1000.
  4. Strickland, D. K. et al. Biochem. 1983, 22, 444. (Abstract)
  5. Bouriotis, V. et al. J. Chrom. 1981, 210, 267.
  6. Cuatrecasas, P.; Illiano, G. Biochem. Biophys. Res. Comm. 1971, 44, 178.
  7. Byrun, Y. et al. J. Biomater., Sci. Polym. Ed. 1994, 6, 1-13.
  8. Thunberg, L. et al. Carbohy. Res. 1982, 100, 393-410. (Abstract)
  9. Johnson, D. A.; Travis, J. Anal. Biochem. 1976, 72, 573-576.
  10. Wichman, A. Biochem. J. 1979, 181, 691-698. (Abstract)
  11. Mosckovitz, R.; Gershoni, J. M. J. Biol. Chem. 1988 263, 1017. (Abstract)
  12. Haidar, M. et al. Glycoconjugate J. 1993, 9, 315.
  13. Mitsakos, A.; Hanisch, F. G. Biol. Chem. Hoppe-Seyler 1989, 370, 239-243. (Abstract)
  14. Salter, D. N. et al. Biochem. J. 1981, 193, 469-476. (Abstract)
  15. Pfeuffer, E. et al. Proc. Natl. Acad. Sci. USA 1985, 82, 3086. (Abstract)
  16. Ruoslahti, E. et al. J. Biol. Chem. 1979, 254, 6054-6059. (Abstract)
  17. Habeeb, A. F. S. A. Biochem. Biophys. Res. Commun. 1981, 100, 1154-1166. (Abstract)
  18. Chiancone, E. et al. J. Chrom. 1992, 604, 117-123. (Abstract)
  19. Landt, M. et al. Biochem. 1978, 17, 915. (Abstract)
  20. Wallis, M. H. et al. Biochem. 1980, 19, 798. (Abstract)
  21. Cayley, P. J. et al. Biochem. 1981, 20, 874-879. (Abstract)
  22. Afting, E. G. Biochem. J. 1981, 197, 519-522. (Abstract)
  23. Niggli, V.; Zurini, M.; Carafoli, E., Methods Enzymol. 1987, 139 791-808. (Abstract)
  24. Dedman, J. R. et al. J. Biol. Chem. 1993, 268, 23025-23030. (Abstract)
  25. Sundberg, L.; Hoglund, S. FEBS Lett. 1973, 37, 70.
  26. Molvig, J.; Baek, L. Scand. J. Immunol. 1987, 26, 611-619. (Abstract)
  27. Issekutz, A. C. J. Immunol. Meth. 1983, 61, 274-281. (Abstract)
  28. Piepkorn, M. W. et al. Arch. Biochem. Biophys. 1980, 205, 315-322. (Abstract)
  29. Wooten, M. W. et al. Eur. J. Biochem. 1987, 164, 461-467. (Abstract)
  30. Van Etten, R. L. et al. Clin. Chem. 1978, 24, 1525. (Abstract)
  31. Lin, M. F. et al. Biochem. 1983, 22, 1055.
  32. Feinstein, G. et al. Eur. J. Biochem. 1974, 43, 569-581.
  33. Peterson, L. M. et al. Biochem. 1976, 15, 2501-2508. (Abstract)
View our listing of Specialty Resins
back to top