|
Gel filtration chromatography seprarates proteins, peptides, and oligonucleotides on the basis of size. Molecules move through a bed of porous beads, diffusing into the beads to greater or lesser degrees. Smaller molecules diffuse further into the pores of the beads and therefore move through the bed more slowly, while larger molecules enter less or not at all and thus move through the bed more quickly. Both molecular weight and three-dimensional shape contribute to the degree of retention. Gel Filtration Chromatography may be used for analysis of molecular size, for separations of components in a mixture, or for salt removal or buffer exchange from a preparation of macromolecules.
Molecular Weight Ranges for Gel Filtration Media
Molecular Weight Ranges for Gel Filtration Media
back to top
Hydrophobic / Lipophilic
Hydroxypropyl beaded dextran (Lipohilic Sephadex®), substituted with long chain alkyl ethers.
| Medium |
Properties / Limitations |
Potential Applications |
| Matrix: hydroxyalkoxypropyl-dextan |
Clean with petroleum ether and acetone |
Separation fatty acids and esters |
Type I: alkyl chain length C11-C14 Type VI: alkyl chain length C15-C18 Type IX: alkyl chain length C15-C18 |
No decomposition over a six month period of continual use |
Fractionation of triglyceride mixtures |
| |
|
Separation of phospholipids, cholesterol and steriods |
Visit our listing of Hydrophobic / Lipophilic
Sephadex is a registered trademark of GE Healthcare.
back to top
Sephacryl®
Cross-linked co-polymer of allyl dextran and N,N’-methylenebisacrylamide. The HR grades are smaller particle sizes with narrower size distributions optimized for more efficient separations with faster flow. Gives fast and easy high-resolution results in both lab and industrial scales.
| Medium |
Fraction Ranges (proteins) |
Properties / Limitations |
Potential Applications |
| Matrix: acrylic (dextran/bisacrylamide copolymer) |
S-100 HR: 1K – 100K |
Clean with 0.2M NaOH, 0.1M HCl, 1.0M HOAc Sterilize by autoclave |
S-100 HR: peptides, small proteins |
25-75 µm S-1000: 40-105 µm |
S-200 HR: 5K – 250K |
Compatible with detergents, chaotropic salts, dissociation agents |
S-200 HR: proteins, some small serum proteins |
| 0.2 MPa |
S-300 HR: 10K – 1500K |
Compatible with organic solvents |
S-300 HR: membrane & serum proteins, monoclonal antibodies |
| pH range: 2-11 |
S-400 HR: 20K – 8000K |
Scaleable Stable bed volume |
S-400 HR: large proteins & macromolecules with extended structures (e.g. proteoglycans, liposomes) |
| |
S-500 HR, S-1000: ND |
Good recoveries |
S-500 HR: large macromolecules, small particles (e.g. plasmids) |
Visit our listing of Sephacryl Matrices
Sephacryl is a registered trademark of GE Healthcare.
back to top
Sephadex®
Beads are prepared by cross-linking dextan with epichlorohydrin. Widely used in industrial processes. Also useful in desalting and buffer exchange.
| Medium |
Fraction Ranges (proteins) |
Properties / Limitations |
Potential Applications |
| Matrix: dextran (crosslinked with epichlorohydrin) |
G-10: <700 |
Clean with 0.2M NaOH Sterilize by autoclave |
G-10: very low MW substances; desalting; buffer exchange; peptides |
C: 100–300 µm M: 50–150 µm F: 20–80 µm SF: 10–40 µm |
G-15: <1500 |
Compatible with detergents, chaotropic salts, dissociation agents |
G-15: low MW compounds; desalting; buffer exchange; peptides |
| SF: 0.016–0.160 MPa |
G-25: 1K –5K |
Interact with aromatic compounds |
G-25: routine desalting; small peptides & proteins; buffer exchange |
pH range: 2–13 (<G-25) 2–10(>G-25) |
G-50: 1.5K – 30K |
Classic Support Swelling 2 – 40X |
G-50: desalting; buffer exchange; standard for process scale; very low nonspecific interactions |
| |
G-75: 3K – 80K |
C: for very crude samples M: for standard lab work F: for preparative lab work SF: for best resolution, low flow rate |
G-100: very low nonspecific interactions |
| |
G-100: 4K – 150K |
Ideal for MW determinations |
|
Visit our listing of Sephadex Matrices
Sephadex is a registered trademark of GE Healthcare.
back to top
Sepharose®
Beaded agarose for fractionation molecules of high molecular weight. Cross-linked beaded agarose is more resistant to denaturing conditions, and thus offers more versatility in the choice of sample buffer and eluent. The approximate % agarose concentration is indicated by the first number of the Stock Number. Often used as a base matrix for coupling affinity ligands or separation of very large molecules and virus particles.
| Medium |
Fraction Ranges (proteins) |
Properties / Limitations |
Potential Applications |
| Matrix: agarose |
6B: 10K – 4000K |
Clean with 0.5M NaOH Cannot be autoclaved Temperature range: 4–40 °C |
6B: polio virus purification; proteins; polysaccharides; affinity support |
6B: 45 – 165 µm 4B: 45 – 165 µm 2B: 60 – 200 µm |
4B: 60K – 20,000K |
Incompatible with oxidizing agents, chaotropic salts, dissociating agents |
4B: tRNAs; membrane proteins; polysccharides; affinity support |
| 0.004 – 0.02 MPa |
2B: 70K – 40,000K |
Broad fraction range |
2B: DNA-protein complexes; viruses; asymmetric molecules; affinity support |
| pH range: 4–9 |
|
Good recoveries |
|
Visit our listing of Sepharose Matrices
Sepharose is a registered trademark of GE Healthcare.
back to top
Superdex®
Composite of cross-linked agarose and dextran. High-resolution with short run times and good recovery.
| Medium |
Fraction Ranges (proteins) |
Properties / Limitations |
Potential Applications |
| Matrix: dextran (covalently bonded to agarose) |
30: <10K |
Clean with 1M NaOH, 0.1M HCl, 1.0M HOAc Sterilize by autoclave |
30: peptides; oligosaccharides; small proteins. Selectivity between Sephadex G-25 & G-50 |
| 24 – 44 µm |
75: 3K – 70K |
Can be used with detergents, chaotropic salts, dissociating agents |
75: high resolution prep separations; wide range of recombinant DNA products. Selectivity similar to Sephadex G-75 |
| 0.5 MPa |
200: 10K – 600K |
Compatible with organic solvents |
200: monoclonal antibodies; high resolution prep separations; contaminants of low MW; good when protein MW is unknown. Selectivity similar to Sephadex G-200 |
| pH range: 3–12 |
|
Scaleable High recoveries Highest selectivity |
|
Visit our listing of Superdex Matrices
Superdex is a registered trademark of GE Healthcare.
back to top
Superose®
Highly cross-linked beaded agarose.
| Medium |
Fraction Ranges (proteins) |
Properties / Limitations |
Potential Applications |
| Matrix: agarose (highly crosslinked) |
12: 1K – 300K |
Clean with 0.2M NaOH, 0.01MHCl, 1.0M HOAc Sterilize by autoclave |
Preparative work: proteins; DNA fragments; polysaccharides |
| 20 – 40 µm |
6: 5K – 5000K |
Compatible with detergents, chaotropic salts, dissociating agents |
|
| 12: 0.7 MPa |
6: 0.4 MPa |
Compatible with organic solvents |
|
| pH range: 2–14 |
|
Some hydrophobic interactions |
|
Visit our listing of Superose Matrices
Superose is a registered trademark of GE Healthcare.
back to top
Toyopearl® HW
Toyopearl is a totally porous, semirigid spherical gel designed for medium and low pressure liquid chromatography. Toyopearl HW gels are synthesized from hydrophilic vinyl polymer containing numberous hydroxyl groups and are composed exclusively of C, H and O atoms. Toyopearl HW is very strong mechanically and can be used at high flow rates.
| Medium |
Fraction Ranges (proteins) |
Properties / Limitations |
Potential Applications |
| Matrix: methacrylate (ethylene glycol/methacrylate copolymer) |
40: <10K |
Clean with surfactant, dissociating agent, acid/base Sterilize by autoclave |
HW-40: very effective with organics; phthalates; cyclodextrins; low MW nucleosides/nucleotides; peptide digests |
C: 50 – 100 µm F: 30 – 60 µm S: 20 – 40 µm |
50: 500 – 80K |
Compatible with organic solvents No saccharide derivative leaching |
HW-50: medium MW molecules; peptides |
| 0.7 MPa |
55: 1K – 700K |
High mechanical strength (5–7 bar) High flow rate |
HW-55: medium MW molecules; RNase digest; tissue plasminogen activator; RNA aggregates; glycoproteins |
| pH range: 2–12 |
65: 40K – 5000K |
Useful for very hydrophobic proteins Stable bed volume |
HW-65: polysaccharides; mRNA; collagenous proteins |
| |
75: 500K – 50,000K |
S: high resolution, high pressure drop, low flow rate C: low resolution, low pressure, high flow rate |
HW-75: large MW molecules; amylose subfractions; plasmid DNA; replace ultracentrigation steps |
Visit our listing of Toyopearl HW Matrices
Toyopearl is a registered trademark of TosoH Corp.
back to top
Ultrogel®
Beaded agarose, free of fines.
| Medium |
Fraction Ranges (proteins) |
Potential Applications |
Matrix: A: agarose AcA: agarose & polyacrylamide |
A6: 25K – 2,400K |
AcA54: 5K – 70K |
Globular proteins |
| 60 – 140 µm |
A4: 55K – 9,000K |
AcA44: 10K – 130K |
|
| pH 3–10 |
A2: 120K – 25,000K |
AcA34: 20K – 350K |
|
| |
|
AcA22: 100K – 1,200K |
|
Visit our listing of Ultrogel Matrices
Ultrogel is a registered trademark of IBF Biotechnics, Inc.
back to Protein Chromatography back to top
|