Sigma-Aldrich
Life Science > Proteomics > Protein Electrophoresis > Western Blotting > Protein MW Markers
Western Blotting
Protein MW Markers
Sigma offers a variety of prestained markers as well as a biotinylated marker for use in Western blotting analysis. Prestained markers contain proteins conjugated to brilliantly colored dyes, and Sigma's biotinylated marker contains biotin conjugated proteins easily detected with a Streptavidin Peroxidase conjugate and color development reagent.
Product #  Product Name MW Range (Da)
SDS7B2 Prestained Molecular Weight Marker (27,000-189,000)
B2787 Biotinylated Molecular Weight Marker (6,500-180,000)
C2242 Chemichrome™ Western Control (8,000 - 220,000)
C2117 Chemichrome™ Ultimate (8,000 - 220,000)
C4861 ColorBurst™ Electrophoresis Marker High (30,000-220,000)
C1992 ColorBurst™ Electrophoresis Marker (8,000-220,000)
C3187 Color Marker Low Range (6,500-45,000)
C6210 Color Marker Ultra-Low Range (1,060-26,600)
ORDERING
SERVICE & SUPPORT
CORPORATE
    Follow us on Twitter!  Join us on Facebook!  Join us on LinkedIn!  Watch our videos on YouTube!
Copyrights © 2010 Sigma-Aldrich Co. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.