Sigma-Aldrich
Life Science > Proteomics > Protein Electrophoresis > Western Blotting > ProteoQwest Kits
Western Blotting
ProteoQwest Kits
ProteoQwest Colorimetric and Chemiluminescent Western Blotting Kits are designed for high sensitivity protein detection directly on Western Blot membranes. The kits include all qualified essential reagents and antibodies used in a Western blot.
ProteoQwest™ Colorimetric Western Blotting Kits:
  • TMB Substrate, for Mouse Monoclonal IgG Antibodies
  • BCIP/NBT Substrate, for Mouse Monoclonal IgG Antibodies
ProteoQwest™ Chemiluminescent Western Blotting Kits:
  • CPS1 Substrate for Mouse Monoclonal IgG Antibodies
  • CPS Substrate for Rabbit Antibodies
  • TMB Substrate for Rabbit Antibodies
ProteoQwest™ Colorimetric Western Blotting Kit, TMB Substrate, for Mouse Monoclonal IgG Antibodies
ProteoQwest Colorimetric Western Blotting Kit, TMB Substrate is designed for high sensitivity colorimetric detection of as little as 0.15 ng. The kit includes all qualified essential reagents and antibodies used in a Western blot specific for mouse monoclonal IgG antibodies. The colorimetric reaction occurs directly on the membrane. No darkroom or film is needed when performing a Western blot with the ProteoQwest Colorimetric Kit. This kit is designed for 25 mini-gel (10 x 10 cm) blots.
Sigma PVDF membrane of ProteoQwest Colorimetric Western Blotting Kit, TMB Substrate
Product #  Product Name Size
PQ0101 ProteoQwest Colorimetric, TMB Substrate 1 kit
back to top
ProteoQwest™ Colorimetric Western Blotting Kit, BCIP/NBT Substrate, for Mouse Monoclonal IgG Antibodies
ProteoQwest Colorimetric Western Blotting Kit, BCIP/NBT Substrate is designed for high sensitivity colorimetric detection for as little as 0.5 ng. This kit includes all qualified essential reagents and antibodies used in a Western blot specific for mouse monoclonal IgG antibodies. This kit is designed for 25 mini-gel (10 x 10 cm) blots.
Sigma PVDF membrane of ProteoQwest Colorimetric Western Blotting Kit, BCIP/NBT Substrate
Product #  Product Name Size
PQ0111 ProteoQwest Colorimetric, BCIP/NBT Substrate 1 kit
back to top
ProteoQwest™ Chemiluminescent Western Blotting Kit, CPS1 Substrate for Mouse Monoclonal IgG Antibodies
ProteoQwest Chemiluminescent Western Blotting Kit with CPS Substrate is designed for high sensitivity chemiluminescent detection of as little as 0.03 ng of protein. It uses the popular Horseradish Peroxidase (HRP) enzyme and includes essential reagents and antibodies used in a Western blot specific for mouse monoclonal IgG. The chemiluminescent reaction occurs directly on the protein immobilized membrane. This kit is optimized for use with either film or chemiluminescent imagers. All components of the ProteoQwest Chemiluminescent Western Blotting Kit have been extensively tested and optimized to ensure superb results. This kit is designed for 12 mini-gel (10 x 10 cm) blots.
Sigma PVDF membrane of ProteoQwest Chemiluminescent Western Blotting Kit, CPS1 Substrate for Mouse Monoclonal IgG Antibodies
Product #  Product Name Size
PQ0201 ProteoQwest Chemiluminescent, CPS-1 Substrate for mouse monoclonal IgG Antibodies 1 kit
CPS-1-60 Chemiluminescent Peroxidase Substrate 60 mL
back to top
ProteoQwest™ Chemiluminescent Western Blotting Kit, CPS Substrate for Rabbit Antibodies
ProteoQwest Chemiluminescent Western Blotting Kit, CPS Substrate is designed for high sensitivity, low background chemiluminescent detection of as little as 0.06 ng of protein. All of the components of this ProteoQwest kit have been extensively tested and optimized. This kit is designed for 12 mini-gel sized (10 x 10 cm) blots. It is possible to use this kit for as many as 24 blots if half the suggested amount of reagents is used.
Product #  Product Name Size
PQ0221 ProteoQwest™ Chemiluminescent, CPS Substrate for Rabbit Antibodies 1 kit
back to top
ProteoQwest™ Chemiluminescent Western Blotting Kit, TMB Substrate for Rabbit Antibodies
The ProteoQwest Colorimetric Western Blotting Kit, TMB Substrate is designed for high sensitvity, low background colorimetric detection of as little as 0.25 ng of protein. The colorimetric reaction occurs directly on the PVDF or nitrocellulose membrane; no darkroom or film is needed. All of the components of this ProteoQwest kit have been extensively tested and optimized. This kit is designed for 25 mini-gel sized blots. (10 x 10 cm)
Product #  Product Name Size
PQ0121 ProteoQwest™ Colorimetric, TMB Substrate for Rabbit Antibodies 1 kit
back to top
back to Western Blotting