|
|||||||||
|
The kits and individual extraction reagents of increasing solubilizing power each of which can generate selective or total protein extracts from cellular samples. The kits include the newest generation of detergents. The protein extractions obtained with each component can be optimized to meet your individual needs. The reducing and alkylating reagents produce protein samples that exhibit improved focusing and decreased streaking in 2D gels. Enough of each component is provided to process a multiople protein samples. For researchers who have optimized an extraction protocol using one chaotropic extraction reagent, each kit reagent is also available as an individual product.
|
Discover how our New Technologies for Proteomic Research can enhance your studies!
|
|||||||
|
|
|||||||||