Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Proteomics > Sample Preparation > Protein Depletion > Seppro® Protein Depletion
Seppro® Protein Depletion
Bioextract ImageOvercome protein sample complexity – Separate with Sigma's Seppro Depletion Technology
Sigma's Seppro Depletion Technology enables removal of interfering highly abundant proteins from a variety of biological samples using the affinity of avian polyclonal IgY antibodies. This removal unmasks low abundance proteins of interest for further investigation.
Products
Human   Remove the top 14 highly abundant proteins in human plasma/serum. Further deplete moderately abundant proteins with the Human SuperMix column.
Mouse   Remove the top 7 highly abundant proteins in mouse plasma/serum. Further deplete moderately abundant proteins with the Mouse SuperMix column.
Rat   Remove the top 7 highly abundant proteins in rat plasma/serum.
Plant   Remove highly abundant RuBisCO from plant samples.
Custom   Interested in a custom Seppro format? Contact Sigma® Life Science about custom formats and sizes to meet your depletion needs. seppro@sial.com
back to top