Sigma-Aldrich
Life Science > Proteomics > Sample Preparation > Protein Depletion > Seppro® Depletion Products
Seppro® Depletion Products
Dig Deeper into the Proteome!
The Seppro depletion technology allows for the removal of several high abundant proteins from a variety of biological samples. The use of Avian polyclonal IgY (Immunoglobulin Yolk) antibodies provides unique and advantageous features that allow highly-specific partitioning of protein mixtures. As a result, previously masked proteins become more accessible for investigation.
  • Depletion from a variety of sample types
  • Spin-Column and LC formats
  • Specific capture of target proteins
Discover The ProteoPrep® 20 Plasma Immunodepletion Technology
Human IgY14 Columns
  IgY14 columns remove the top 14 highly abundant proteins in human plasma/serum.
Human Supermix System
  Human Supermix columns remove other moderately abundant proteins from human plasma/serum when used in conjunction with
  the IgY14 system.
IgY Mouse-Specific Columns
  IgY mouse specific coulmns remove 7 highly abundant proteins from mouse plasma/serum.
IgY Rat-Specific Columns
  IgY rat specific coulmns remove 7 highly abundant proteins from rat plasma/serum.
IgY RuBisCO Columns
  Specifically removes Rubisco protein from plant extract.
FAQs