Extraction Kits Reagents
ProteoPrep® 20 LC
Discover Protein Biomarkers Faster
ProteoPrep 20 Plasma Immunodepletion LC Column (PROT20LC)
By removing 20 highly abundant proteins (representing 97% of the proteome) at 98% efficiency, less fractionation of the true plasma proteome is required; focusing on your protein biomarkers becomes faster and simpler!
- Reduce sample complexity and increase efficiency
- Reveal proteins previously masked
- Enrich proteins of interest up to 20-fold
Designed for use with LC instruments, the ProteoPrep 20 LC column accommodates a sample load of 100 µL plasma or serum and can be used up to 100 times.
Read the press release:
Biomarker Discovery Simplified with Innovative Technology from Sigma-Aldrich®
Components
- 1 Depletion Column
- 10X Equilibration Buffer
- 10X Elution Buffer
- Kathon (for long-term storage)
- Collection Tubes
- Spin Filters
Ordering Information
| Product No. |
Product Name |
Package Size |
| PROT20LC |
ProteoPrep 20 Plasma Immunoaffinity LC Column |
1 kit |
ProteoPrep 20 Plasma Immunodepletion Technology
| The ProteoPrep 20 Technology depletes the following proteins: |
Albumin IgGs Transferrin Fibrinogen IgAs α-2-Antitrypsin IgMs α-1-Antitrypsin Complement C3 Haptoglobulin |
Apolipoprotein A1 Apolipoprotein A2 Apolipoprotein B Acid-1-Glycoprotein Ceruloplasmin Complement C4 Complement C1q IgDs Prealbumin Plasminogen |
The ProteoPrep 20 technology offers the highest depletion of highly abundant plasma proteins available today and specifically removes 20 highly abundant proteins (representing 97% of the proteome) to reveal a new layer of the plasma proteome.
| Spot No. |
Protein Identification |
1 2 3 4 5 6 7 8 9 10 11 12 13 14 15 16 17 18 19 20 21 22 |
Alpha-1-B-glycoprotein Hemopexin (Beta-1B-glycoprotein) Alpha-2-HS-glycoprotein (Fetuin A) Vitamin D Binding Protein Pigment Epithelium-derived factor Carboxypeptidase N (Kininase 1) Complement Factor H fragment Apolipoprotein E Ficolin 3 (Hakata antigen) PK-120 (Plasma Kallikrein-sensitive Glycoprotein) Apolipoprotein A-IV Glutathione Peroxidase 3 Tetranectin (Plasminogen Binding Protein) Serum amyloid A (SAA) Kininogen Complement Factor H-related protein 2 Inter-alpha-trypsin inhibitor H1 Vitronectin Geltain-Binding Protein Antithrombin III Cluterin (Apolipoprotein J) Inter-alpha-trypsin inhibitor H4 |
22 Unique Proteins Revealed with ProteoPrep 20
The ProteoPrep 20 Plasma Immunodepletion Kit specifically removes 20 highly abundant proteins (representing 97% of the proteome) at 99% efficiency, revealing a new layer of highly abundant proteins. Initially, the gel image above appears to be similar to an undepleted sample, but identifications via mass spectrometry indicate that the visible proteins are not members of the list of twenty high abundance proteins specifically removed. They are, indeed, unique to a sample depleted with the ProteoPrep 20 technology.
References
Resources
Related Products
ProteoPrep 20 Plasma Immunodepletion Kit
ProteoPrep 20 spin columns specifically remove 20 of the most abundant proteins from human plasma or serum to reveal previously masked protein biomarkers. Additionally, this facilitates a 20-fold increase in the relative amount of revealed proteins that can be subsequently studied and analyzed. Designed for use with bench top centrifuges, ProteoPrep 20 spin columns can accommodate a sample load of 8 µL plasma or serum and can be used up to 100 times each. The ProteoPrep 20 Plasma Immunodepletion Kit contains 3 spin columns. The ProteoPrep 20 Sample Kit contains 1 spin column.
Components
- Depletion Spin Column(s)
- 10X Equilibration Buffer
- 10X Elution Buffer
- Kathon (for long-term storage)
- Collection Tubes
- Spin Filters
- Syringes
- Luer Lock Caps
| Product No. |
Product Name |
Package Size |
| PROT20 |
ProteoPrep 20 Plasma Immunodepletion Kit |
1 kit (3 columns) |
| PROT20S |
ProteoPrep 20 Plasma Immunodepletion Sample Kit |
1 kit (1 column) |
ProteoPrep Blue Albumin & IgG Depletion Kit
ProteoPrep Blue Albumin Depletion Kit is designed to specifically remove albumin and IgG from 25 samples of human serum (25 µl to 100 µl) in preparation for two-dimensional electrophoresis (2DE). Albumin-depleted serum samples generated by this kit are in urea rather than salt-based buffers with little dilution, eliminating the need to precipitate the sample prior to performing 2DE.
Components
- ProteoPrep Blue Albumin and IgG Depletion Medium—a 10-ml suspension containing 60% packed medium
- ProteoPrep Blue Equilibration buffer—1 bottle reconstitutes to 50 ml, pH 7.8
- Protein Extraction Reagent Type 4—1 bottle reconstitutes to 23 ml, pH 10.4
- 25 Spin columns
- 75 Collection Tubes
| Product No. |
Product Name |
Package Size |
| PROTBA |
ProteoPrep Blue Albumin & IgG Depletion Kit |
1 kit |
back to top