IgY Rat-Specific Columns
Immunoaffinity partitioning of highly-abundant proteins (HAP) has proven to be one of the most effective approaches for overcoming the wide dynamic range of plasma protein concentration, as well as enabling the detection of low-abundant proteins (LAP). Avian polyclonal IgY (Immunoglobulin Yolk) antibodies have unique and advantageous features that allow highly-specific partitioning of protein mixtures. Sigma offers a novel Seppro® IgY-R7 column system, which takes you deeper into the rat plasma/serum proteome.
Proteins Partitioned by IgY-R7 columns: 
  • Rat serum albumin
  • IgG
  • Fibrinogen
  • Transferrin
  • IgM
  • Haptoglobin
  • alpha1-Antitrypsin
Columns are available in the following formats:
Product Name Column Use Rat plasma/serum Product #
IgY-R7 Spin Column Kit
Includes 2 spin columns and reagents.
Good for 200 samples.
100 times 15µl SEP130 
IgY-R7 LC-10 Column Kit
Includes 1 HPLC column and reagents.
100 times 200µl SEP120