|
|||||||||||||||||||||||||||||||||||||||||||
|
|||||||||||||||||||||||||||||||||||||||||||
|
Immunoaffinity partitioning of highly-abundant proteins (HAP) has proven to be one of the most effective approaches for overcoming the wide dynamic range of plasma protein concentration, as well as enabling the detection of low-abundant proteins (LAP). Avian polyclonal IgY (Immunoglobulin Yolk) antibodies have unique and advantageous features that allow highly-specific partitioning of protein mixtures. Sigma® offers a novel Seppro® IgY14 columns system, which takes you deeper than ever into the plasma proteome. IgY14 columns remove the top 14 HAP. Proteins Partitioned by IgY-14 Microbeads:
Our IgY14 systems are available in the following formats (suitable for your volumes):
Human SuperMix System Immunoaffinity fractionation of highly-abundant proteins (HAP) has proven to be one of the most effective approaches for overcoming the wide dynamic range of protein concentration, as well as enabling the detection of low abundant proteins (LAP). Avian polyclonal IgY (Immunoglobulin Yolk) antibodies have unique and advantageous features that allow for highly-specific fractionation of a complex protein mixture. The IgY14 column can effectively remove 14 HAP from plasma. The next level of abundant proteins, moderately abundant proteins (MAP), becomes an obstacle to access LAP, where the majority of biologically interesting and clinically important biomarkers reside. Therefore, isolation of MAP is a new challenge for effectively and accurately detecting and analyzing LAP. To tackle this challenge, the SuperMix column was developed to separate MAP from LAP. The SuperMix System was developed by immunizing chickens with a flow-through fraction of IgY-12 or IgY14 column and constructing the column with affinity-purified IgY antibodies against the flow-through proteins of IgY-12 or IgY14. SuperMix Fractionation Process The Human SuperMix column is then used in conjunction with the IgY14 column to provide superior depletion. ![]() SuperMix Results As shown below, the SuperMix column was applied for fractionating the flow-through fraction of IgY14, which resulted in a bound/eluted fraction (designated MAP fraction) and the flow-through fraction (designated LAP fraction). Unfractionated and serial-fractionated samples using IgY14 and SuperMix columns were analyzed by SDS-PAGE and 2DE. Our data demonstrate that SuperMix columns specifically and reproducibly remove a group of proteins, which are the main abundant proteins in the flow-through fraction of IgY14. ![]()
Recommended Set-Up:
|
|||||||||||||||||||||||||||||||||||||||||||
|
|
|||||||||||||||||||||||||||||||||||||||||||