Sigma-Aldrich
Life Science > Proteomics > Sample Preparation > Protein Depletion > Seppro® Protein Depletion > IgY RuBisCO Columns
IgY RuBisCO Columns
Similar to the situation of plasma or serum proteomics, plant proteomics study also faces the challenges of specifically removing the HAP from plant protein extract in order to analyze or detect the LAP. 
RuBisCO (Ribulose-1,5-bisphosphate carboxylase/oxygenase) is the most abundant protein in plants, and it may be the most abundant protein on Earth. RuBisCO is an enzyme that is used in the Calvin cycle to catalyze the first major step of carbon fixation, a process by which the atoms of atmospheric carbon dioxide are made available to organisms in the form of energy-rich molecules such as sucrose. RuBisCO, while being the key enzyme in photosynthetic carbon assimilation in green leaves, is the main obstacle in plant proteomics. It constitutes about 40% of the total protein mass in green leaves, thus interfering with proteomics studies such as LC-MS/MS and 2D-gel electrophoresis. To meet the needs for specifically and effectively separating RuBisCO from other plant proteins, the novel immunoaffinity matrix based on the IgY antibodies cross-linked to microbeads, which specifically removes RuBisCO protein from plant extract was developed.
Seppro RuBisCO Column Features:
  • High-efficiency RuBisCO removal
  • High column capacity
  • Successfully applied for processing a variety of plant samples
Seppro RuBisCO x-ray diffraction model
X-ray diffraction based model showing
Spinach RuBisCO in complex with
2-carboxyarabinitol 2 bisphosphate
and calcium.
Cellar NA, Kuppannan K, Langhorst ML, Ni W, Xu P, Young SA. (2007). Cross species applicability of abundant protein depletion columns for ribulose-1,5-bisphosphate carboxylase/oxygenase. J Chromatogr B Analyt Technol Biomed Life Sci. Epub.
Product Name Use Capacity of RuBisCO protein Product #
Anti-RuBisCO Spin Columns
Includes 2 spin columns.
100 times 0.2 mg SEP070
Anti-RuBisCO HPLC LC-2 column
Includes 1 HPLC column.
100 times 1 mg SEP080