|
|||||||||||||||||||||
|
|||||||||||||||||||||
|
Immunoaffinity fractionation of highly-abundant proteins (HAP) has proven to be one of the most effective approaches for overcoming the wide dynamic range of protein concentration, as well as enabling the detection of low abundant proteins (LAP). Avian polyclonal IgY (Immunoglobulin Yolk) antibodies have unique and advantageous features that allow for highly-specific fractionation of a complex protein mixture. The IgY14 column can effectively remove 14 HAP from plasma. The next level of abundant proteins, moderately abundant proteins (MAP), becomes an obstacle to access LAP, where the majority of biologically interesting and clinically important biomarkers reside. Therefore, isolation of MAP is a new challenge for effectively and accurately detecting and analyzing LAP. To tackle this challenge the SuperMix column was developed to separate MAP from LAP. The SuperMix System was developed by immunizing chickens with a flow-through fraction of IgY-12 or IgY14 column and constructing the column with affinity-purified IgY antibodies against the flow-through proteins of IgY-12 or IgY14. SuperMix Fractionation Process ![]() SuperMix Results ![]()
Recommended Set-Up:
|
|||||||||||||||||||||
|
|
|||||||||||||||||||||