Sigma-Aldrich
Life Science > Proteomics > Protein Structural Analysis > X-Ray Crystallography > Basic Crystallography Kit
X-Ray Crystallography
Basic Crystallography Kit
Crystallization Plates | Membrane Protein Kit | Optimization Reagents | Renaturation Kit
Features: Benefits:
Variety of proven precipitants Allows rapid and simplified screening across a broad range
High-purity reagents Ensures reliable, reproducible results
Sterile solutions Avoids waste of precious sample
Conveniently-packaged ready-made solutions Reduces waste, allows easy handling
Individual solutions and reagents are available Simplifies scale-up of crystallization following determination of optimal conditions
Each screening kit consists of 48 or 50 different crystallization conditions. 10 mL of each compound are included in the kit; individual compounds are available in 100 mL package size.
Basic Crystallography and Extension Kits
Crystallization Kit for Proteins for Automated Handling
Cryo Kit
Low Ionic Kit
PEG Grid Screen Kit
Membrane Protein Kit
Crystallization Kit for Protein-Protein Complexes
DNA Crystallization Kit
RNA Crystallization Kit
crystal-image
Basic Crystallography and Extension Kits
The Basic Crystallography Kit is a rapid, empirical, and efficient screening method for determining the solubility and optimal starting conditions for the crystallization of biological macromolecules. Based on known and published data for various proteins, the Basic Kit uses a sparse matrix method and a minimal amount of protein sample to explore a broad range of buffers, pH, and precipitants (Jancarik, J. and Kim, S.H., J. Appl. Cryst., 24, 409-411, 1991).
Basic Crystallography Kit
The Extension Kit substantially increases the screening range of the Basic Crystallography Kit by providing a broader range of organic solvents, PEGs, and additives (divalent ions and stabilizers).
Both the Basic and Basic Extension Kit are based on the "salting-out" principle: that increasing the concentration of the precipitating agent depletes the macromolecules of bound water and leads to their precipitation and crystallization.
Product #  Product Name Size
82009 Basic Crystallography Kit 1 kit
70437 Basic Extension Kit 1 kit
Screening Parameters Basic Kit Extension Kit
pH Range 4.6 to 8.5 4.6 to 9.0
Buffers acetate, tartrate, phosphate, Tris, citrate, HEPES, imidazole, formate, and cacodylate MES, Bicine, Tris, citrate, HEPES, acetate
Precipitating Salts* tartrate; phosphate; ammonium and lithium sulfate; magnesium and calcium chloride; magnesium, ammonium, sodium, zinc and calcium acetate; sodium citrate; sodium and magnesium formate tartrate; phosphate; magnesium and sodium chloride; sodium acetate; sodium citrate; ammonium formate; lithium and ammonium sulfate; imidazole; CTAB
Precipitating Organic Solvents* MPD, 2-propanol MPD, 2-propanol, ethylene glycol, dioxane, ethanol, 1,6-hexanediol, tert-butanol, glycerol
PEGs* PEG 400, 1500, 4000, and 8000 PEG 400, 6000, 1000, 8000, 10000, and 20000, PEG MME 550, 2000, 5000, and 2000, Jeffamine M-600
Additives   Co2+, Cd2+, Fe3+, Ni2+, and Zn2+ ions, dioxane, ethylene glycol, polyethyleneimine
* As sole precipitant and/or as combinations
Helpful Notes:
Seeding at subcrystallization conditions may produce larger or more regular crystals.
Flow Diagram for Basic Crystallography Kit & Extension Kit
Basic Kit Datasheet including composition (142 kb PDF)
Extension Kit Datasheet including composition (110 kb PDF)
back to top
Crystallization Kit for Proteins for Automated Handling
This Kit is based on our Crystallization Kit for Proteins and the Crystallization Extension Kit for Proteins, which have been combined. The combined screen is delivered as Deepwell-Plate including 1 mL of each compound. This format makes it easy to fill crystallization plates either by automated liquid handling or with the help of 8-channel-pipets. The convenience of this kit saves a lot of time.
Proteins for Automated Handling
Product #  Product Name Size
56783 Crystallization Kit for Proteins for Automatic Screening 1 kit
Helpful Notes:
Flow Diagram for Basic Crystallography Kit & Extension Kit
Extension Kit Datasheet including composition (129 kb PDF)
back to top
Cryo Kit
The crystallization Cryo Kit is a rapid, empirical, and efficient screening method to determine the solubility and optimal starting conditions for the crystallization of biological macromolecules in the presence of a cryoprotectant (primarily glycerol). The screening range and probing ability of the Cryo Kit are comparable to those of the Basic Kit. A cryoprotectant is added to freeze crystals in an amorphous glass at -173 °C. The Cryo Kit is also based on the "salting-out" principle.
Protein Crystallization Cryo Kit
Product #  Product Name Size
75403 Protein Crystallization Cryo Kit 1 kit
Screening Parameters Basic Kit
pH Range 4.6 to 8.5
Buffers Tris, citrate, HEPES, tartrate, phosphate, cacodylate, acetate, imidazole, and formate
Precipitating Salts* tartrate; sodium, potassium, and ammonium phosphate; magnesium and calcium chloride; sodium, magnesium, zinc, calcium, and ammonium acetate; sodium citrate; sodium formate; lithium and ammonium sulfate; calcium and sodium chloride
Precipitating Organic Solvents* MPD, 2-propanol, glycerol
PEGs* PEG 400, 4000, and 8000
* As sole precipitant and/or as combinations
Helpful Notes:
Crystals cracking upon freezing indicates the need for a higher glycerol concentration or another cryoprotectant.
Due to the presence of a cryoprotectant, the Cryo Kit has an increased probability to produce false crystals of low molecular weight components, as compared to the Basic Kit. We recommend testing the nature of initial crystals at an early stage of experimentation by using controls without protein, by staining, or by X-ray defraction.
This kit consist of 50 ready-to-use solutions reflecting different crystallization conditions. When working with standard multiwell plates you may prefer to use just 48 different conditions. In that case we recommend omitting reagents 31 and 48 or two other reagents of your choice.
Flow Diagram for Protein Crystallization Cryo Kit
Cryo Kit Datasheet including composition (117 kb PDF)
back to top
Low Ionic Kit
The Crystallization Low Ionic Kit for proteins is based on a screening protocol for monoclonal antibodies, which has been proven to be an effective screening method for soluble protein. The high-efficiency of this kit can be further improved by pre-determining the isoelectric point (pl) of the subject macromolecule followed by screening within a range at or near that value (within 2-3 pH units of the pl).
Low Ionic Kit
The Low Ionic Kit is based on the "salting-in" principle: that decreasing ion concentrations leads to a loss of electrostatic screening of charges on the macromolecules by low molecular weight ions. This loss of screening is compensated by the mutual screening of opposite charges on neighboring macromolecules. This leads to decreased macromolecular solubility and subsequent precipitation or crystallization.
Product #  Product Name Size
86684 Low Ionic Kit 1 kit
Screening Parameters Basic Kit
pH Range 3.0 to 10.0 (steps of 0.5 pH units)
Buffers citrate, MES, Tris, imidazole, HEPES, glycine, Bis-tris
PEGs* PEG 3350 (4 to 28%)
* As sole precipitant
Helpful Notes:
The salting-in method of crystallography is extremely pH-sensitive. To further reduce the time, effort, and amount of sample needed, we recommend determination of the isoelectric point (pI) of the subject macromolecule followed by screening within two to three pH units of the pI.
Larger crystals may be grown by using seeding in salting-in experiments.
Small drops should be used in vapor-diffusion salting-in as drops will increase in size during the experiment.
Flow Diagram for Low Ionic Kit
Low Ionic Kit Datasheet including composition (103 kb PDF)
back to top
PEG Grid Screen Kit
The Crystallization PEG Grid Screening Kit for Proteins is a screening method for the determination of initial crystallization conditions for biological macromolecules like proteins.
The kit displays a range of conditions based on PEG 600 or PEG 6000 as precipitants and covers various buffers with pH values from pH 4.0 to pH 9.0. PEG is the most widely used type of precipitant, and size and concentration of PEG is critical for crystallization. On the other hand, pH is one of the mayor determinants for crystallization. The use of Crystallization PEG Grid Screening Kit enables determination of suitable conditions regarding these two crucial aspects. Further optimization may include the use of different types and concentrations of various salts.
Product #  Product Name Size
36436 PEG Grid Screen Kit 1 kit
Screening Parameters Basic Kit
pH Range 4.0 to 9.0
Buffers BICINE, HEPES, MES, Tris, citrate
PEGs* PEG 600 and 6000, 5-30%
* As sole precipitant
Helpful Notes:
Crystallization PEG Grid Screening Kit Technical Bulletin (89 kb PDF)
back to top
Membrane Protein Kit
The Crystallization Basic Kit for Membrane Proteins is a rapid, empirical, and efficient screening method to determine the best conditions for crystallization of hydrophobic macromolecules (primarily membrane proteins) in an empirical way. Based on known or published data for various proteins, broad ranges of buffers, pH, and precipitants in combination with a micelle-forming detergent are explored using the sparse matrix method with a minimal amount of protein. The crystallization principle of the kit is based on the screening of the hydrophilic, charged parts of the macromolecules by the binding of the detergent to hydrophobic areas of the macromolecules and subsequent reduction of their solubility. Detergent mediated contacts then lead to precipitation or crystallization of the screened molecules.
Product #  Product Name Size
73513 Membrane Protein Kit 1 kit
Screening Parameters Basic Kit
pH Range 4.6 to 8.5
Buffers Tris, citrate, ADA, HEPES, acetate
Precipitating Salts* Sodium and potassium tartrate; sodium, potassium and ammonium phosphate; sodium and magnesium chloride; sodium and zinc acetate; sodium citrate; lithium, magnesium and ammonium sulfate
Precipitating Organic Solvents* MPD, 2-propanol
PEGs* PEG 400, 4000, and 6000
Detergent** stock solution of choice
Helpful Notes:
We recommend using detergents that form small micelles, followed by use of detergents, which form increasingly larger micelles only as needed.
The ternary system of sample, detergent, and precipitant is a delicate system where small parameter changes can dramatically change interactions between sample-sample, sample-solvent, sample-detergent and detergent-detergent. Corresponding effects on macromolecular solubility and crystallization should be expected.
Temperature is an important variable in all detergent-driven crystallization experiments.
Streak-seeding can produce larger crystals from microcrystalline precipitates.
This kit consists of 50 ready-to-use solutions reflecting different crystallization conditions. When working with standard multiwell plates you may prefer to use just 48 different conditions. In this case we recommend omitting reagents 43 and 47, or two reagents of your choice.
back to top
Crystallization Kit for Protein-Protein Complexes
Crystallization Kit for protein-protein complexes is a rapid empirical screening method to determine the best conditions for the crystallization of such complex protein structures. This kit can also be used for determining the solubility of a certain macromolecule, screening a wide range of pH and different types and concentration of precipitants.
Protein complexes play a crucial role for studies of biological function. Based on own experience and a survey of successful crystallization conditions a specific screen was developed by S. Radaev and P.D. Sun (Applied Crystallography, 35, 674-676, 2002). This kit reflects the most probable conditios for crystallization of protein-protein complexes.
Product #  Product Name Size
02528 Crystallization Kit for Protein-Protein Complexes 1 kit
Screening Parameters Basic Kit
pH Range 5.5 to 8.5
Buffers HEPES, MES, MOPS, Tris, cacodylate, citrate, phosphate
Salts Calcium and magnesium acetate; sodium and magnesium chloride; ammonium and lithium sulphate; potassium, sodium tartrate
Organic Solvents 2-propanol
PEGs PEG 400, 1000, 1500, 2000, 3350, 4000, 6000 and 8000, MPEG 550, 2000 and 5000
Helpful Notes:
Compared to the individual proteins, protein-protein complexes often require less concentrated precipitatns to crystallize. If data for individual compounds is available, this may give an indication, where one should mainly expect crystals to grow.
Crystallization conditions for oligomeric proteins like photosynthesis centres are highly individual and may need a customized screening procedure.
Crystallization Kit for Protein-Protein Complexes Technical Bulletin (92 kb PDF)
back to top
DNA Crystallization Kit
The Crystallization Kit for DNA is an empirical screening method for the direct determination of suitable crystallization conditions for nucleic acids. The kit is developed according to the described method of Nowakowski, et al. (Acta Cryst., D55, 1885-1892, 1999.) The solution and crystallization conditions are empirically derived based on known or published crystallization conditions of nucleic acids, or DNA-protein complexes, especially of DNA-processing enzymes. Crystallization of protein-nucleic acid complexes is an area of increasing interest.
The Crystallization Kit for nucleic acids consists of 48 different formulations covering a range of different buffers (pH 5.5-8.0), different types of precipitants, esp. ethanol, 2-propanol, PEG, and inorganic salts including magnesium salts.
Product #  Product Name Size
80701 DNA Crystallization Kit 1 kit
Screening Parameters Basic Kit
pH Range 5.5 to 8.0
Buffers HEPES, Tris, cacodylate, sodium succinate
Salts Calcium acetate; calcium, lithium, magnesium, potassium and sodium chloride; ammonium, copper and lithium sulphate; sodium tartrate; cobalt hexamine
Organic Solvents Ethanol, MPD, 2-propanol
PEGs PEG 400, 4000, and 8000
Polyamines Spermine, spermidine
Helpful Notes:
For the crystallization of protein-DNA complexes length of DNA plays a mayor role. If initial screening does not yield sufficient crystallization, consider the possibility to change length of DNA.
Crystallization Kit for DNA Technical Bulletin (93 kb PDF)
back to top
RNA Crystallization Kit
The Crystallization Kit for RNA is an empirical screening method for the direct determination of suitable crystallization conditions for RNA and DNA oligomers. Thus it provides a completion of our crystallization kit for DNA. The kit is developed according to the described method of Baeyens, Jancarik and Holbrook (Acta Cryst., D50, 764-767, 1994).
The Crystallization Kit for RNA consists of 48 different formulations covering a range of different buffers (pH 6.0-8.0), different types of precipitants (esp. PEG 400 and MPD), and inorganic salts including magnesium salts.
Product #  Product Name Size
18839 RNA Crystallization Kit 1 kit
Screening Parameters Basic Kit
pH Range 6.0 to 8.0
Buffers HEPES, Tris, cacodylate, sodium succinate
Salts Sodium acetate; ammonium, barium, cadmium, calcium, cobalt, cobalt hexamine lithium, magnesium, manganese, potassium, ruthenium, samarium, sodium and zink chloride; putrescin di-hydrochloride
Organic Solvents Acetone, dioxane, tert-butanol, ethanol, MPD, 2-propanol
PEGs PEG 400, 600, 1000, 4000, and 8000
Polyamines and other compounds Spermine, spermidine, arginine amide
Helpful Notes:
Highly purified RNA is much easier to crystallize than heterogeneous (impure) RNA. Typically, it is much more difficult to purify larger RNA fragments to homogeneity than smaller ones.
In many case, questions can be answered by the investigation of certain fragments of natural RNA. Thus the use of smaller RNA fragments may be a possibility to improve crystallization.
Crystallization and crystal structure can also be influenced by other methods of RNA design, e.g. sequence variation, interhelical stacking, formation of protein-RNA complexes, or incorporation of modiied nucleotides.
Crystallization Kit for RNA Technical Bulletin (99 kb PDF)
back to top
back to X-Ray Crystallography