Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
X-Ray Crystallography
High Throughput Kits
Crystallization Kit for Proteins for Automated Handling:
This Kit is based on our Crystallization Kit for Proteins (no. 82009) and the Crystallization Extension Kit for Proteins (no. 70437), which have been combined.
The combined screen is delivered as Deepwell-Plate including 1 mL of each compound. This format makes it easy to fill crystallization plates either by automated liquid handling or with the help of 8-channel-pipets. The convenience of this kit saves a lot of time.
Product #56783
Product #  Product Name Size
56783 Crystallization Kit for Proteins for Automatic Screening 1 kit
Additional Information:
Flow Diagram for Basic Crystallography Kit & Extension Kit
Datasheet including composition (129 kb PDF)
Handling and Procedures
back to top
High Throughput Crystallization Low Ionic Kit:
The Kit is based on our Crystallization low ionic strength kit for proteins (no. 86684) and the Crystallization PEG grid screening kit for proteins (no. 36436). The kit displays a range of conditions based on PEG 600, PEG 3350, or PEG 6000 as precipitants and covers various buffers with pH values from pH 3.0 to pH 10.0.
The combined screen is delivered as Deepwell-Plate including 1 mL of each compound. This format makes it easy to fill crystallization plates either by automated liquid handling or with the help of 8-channel-pipets. The convenience of this kit saves a lot of time.
Product #02828
Product #  Product Name Size
19554 High Throughput Crystallization Low Ionic Kit 1 kit
Additional Information:
Datasheet including composition (129 kb PDF)
Handling and Procedures
back to top
High Throughput Crystallization Kit for Membrane Proteins and Protein Complexes:
The Kit is based on our Crystallization kit for membrane proteins (no. 73513) and the Crystallization kit for protein complexes (no. 02525).
The combined screen is delivered as Deepwell-Plate including 1 mL of each compound. This format makes it easy to fill crystallization plates either by automated liquid handling or with the help of 8-channel-pipets. The convenience of this kit saves a lot of time.
Product #30653
Product #  Product Name Size
30653 High throughput crystallization kit for membrane proteins and protein complexes 1 kit
Additional Information:
Handling and Procedures
back to top
High Throughput Crystallization Screening Kit for Nucleic Acids:
The Kit is based on a combination of conditions from our crystallization kit for DNA (no. 80701) and our crystallization kit for RNA (no. 18839).
The combined screen is delivered as Deepwell-Plate including 1 mL of each compound. This format makes it easy to fill crystallization plates either by automated liquid handling or with the help of 8-channel-pipets. The convenience of this kit saves a lot of time.
Product #02828
Product #  Product Name Size
02828 High throughput crystallization screening kit for nucleic acids 1 kit
Additional Information:
Handling and Procedures
back to top
High Throughput Crystallization Additives Kit:
The additive kit provides 96 different solutions of various additives, including organic solvents, salts, amino acids, detergents and other compounds.
This kit provides an easy tool for rapid screening of additives. The kit includes 100 µl of each pre-formulated compound and comes in a deepwell block, to allow convenient handling by either automated liquid handling or multi channel pipets.
Product #38428
Product #  Product Name Size
38428 High Throughput Crystallization Additives Kit 1 kit
Additional Information:
Handling and Procedures
back to top
Handling and Procedures
Before use, centrifugation of the deepwell Block with a swing-out rotor centrifuge is suggested, to remove solutions from the cover foil. Centrifugation prevents cross contamination while opening the plate. Otherwise drops under the cover foil may be shaken down. The film should be very gently peeled from the plate.
Figure_01
First open the strap and turn it down to the surface of the deep well plate.
Figure_02
Then smoothly pull the strap in horizontal direction so that the aluminium foil moves parallel to the plate surface to avoid contamination of the wells by drops hanging on the foil.
Instead of peeling the film, it can also be left intact and pierced to take out reagent.
The following procedure describes the use of this kit with the Sitting Drop Vapor Diffusion method.
For manual handling, either a 8 or 12 channel pipet can be used to pipet the recommended volume (50-100 µl) into the reservoirs of a 96 well crystallization plate. Pipet tips should be changed for each set of conditions. But several crystallization plates may be prepared at once (row or column wise) to save tips.
Transfer 50 nl to 2 µl of crystallization reagent from reservoir to the sitting drop well (for each well). Again a 8 or 12 channel pipet may be used. Pipet tips should be changed for each well or set of wells.
Pipet same volume of sample (50 nl to 2 µl) into each sitting drop well. Mixing can be performed by gently aspirating and dispensing the drop several times.
Seal the crystallization plate as you usually do or according to the suppliers recommendations. Seal the deepwell block using foils or cover mats (for short term storage). You may use the enclosed aluminium foils. Simply pull the aluminium foil from the protective film and put the foil on the plate by pressing it onto the surface with evenly spread pressure. Take care that the foil is flush with one side of the plate and overhanging on the other side. To simplify the reopening of the kit put a piece of adhesive tape on the underside of the overhanging aluminium foil.
Please note: When kits are sealed manually we highly recommend to store the kits in upright position at 4°C in the dark.
Sample solubility is also temperature dependent. Allthough most crystallizations have been achieved at room temperature, in many cases different temperatures have led to success. Comparison of results of screening at two different temperatures (4°C and room temperature) helps determine the magnitude of temperature effects on sample solubility. Temperature may be an important parameter in the optimization procedure.
Due to the standard format of deepwell-blocks, they are suitable for any type of standard liquid handler (working with single tip or needle up to 96 parallel channels).
back to top