Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
X-Ray Crystallography
Optimization Reagents
Crystallization Kits | Crystallization Plates | Membrane Protein KitRenaturation Kit
Optimization Reagents
For further optimization of crystallization conditions we are introducing a wide range of single component solutions capable for modification of existing screening kits as well as preparation of customized optimization screening conditions. These solutions are made up from our ultra pure reagents (Biochemika ultra), and highest quality water. They are sterilized and batch-to-batch consistency is subject of extensive analysis. We will continue to add more solutions for optimization of crystallization. Based on our wide range of ultra pure reagents for crystallization and extensive experience for production of formulated reagents we are also ready to prepare customized reagents.
Optimization Reagent Product #  Size
Polyethylene glycol 1000 solution 50% 76293 100 mL, 250 mL
Polyethylene glycol 1500 solution 50% 73034 100 mL, 250 mL
Polyethylene glycol 2000 solution 50% 83918 100 mL, 500 mL
Polyethylene glycol 3350 solution 50% 83272 100 mL, 500 mL
Polyethylene glycol 4000 solution 50% 89782 100 mL, 500 mL
Polyethylene glycol 5000 monomethyl ether Solution 50% 90364 100 mL, 250 mL
Polyethylene glycol 8000 solution 50% 83271 100 mL, 500 mL
Polyethylene glycol 10000 solution 50% 84184 100 mL, 250 mL
Polyethylene glycol 20000 solution 50% 87006 100 mL, 250 mL
Ammonium chloride solution 5M 88163 250 mL, 1 L
Ammonium citrate dibasic Solution 2.5M 85669 100 mL, 500 mL
Ammonium formiate 10M 78314 100 mL, 500 mL
Ammonium phosphate dibasic Solution 2.5M 70705 100 mL, 500 mL
Ammonium phosphate monobasic Solution 2.5M 75568 100 mL, 500 mL
Ammonium sulfate solution 3.2 M 83267 100 mL, 250 mL, 500 mL
Ammonium tartrate dibasic Solution 2M 70631 100 mL, 500 mL
BICINE solution 1M 83263 100 mL, 250 mL, 500 mL
HEPES Sodium salt buffer solution 1M 70216 100 mL, 500 mL
HEPES solution 1M 83264 100 mL, 500 mL
1,6-Hexanediol Solution 6M 88571 100 mL
Imidazol solution 1M 68268 100 mL, 500 mL
Lithium acetate Solution 5M 73216 100 mL, 500 mL
Lithium chloride solution 10M 83268 100 mL, 500 mL
Lithium citrate tribasic Solution 1.5 M 90267 100 mL, 500 mL
Lithium sulfate Solution 2M 82348 100 mL, 500 mL
Magnesium chloride solution 2M 68475 100 mL, 250 mL
Magnesium formiate 0.5M 73068 100 mL, 500 mL
MES buffer Solution 0.5M 76039 100 mL, 500 mL
Potassium acetate solution 5M 95843 100 mL, 250 mL
Potassium chloride Solution 4M 87526 100 mL, 500 mL
Potassium citrate tribasic solution 2.5M 89306 100 mL, 500 mL
Potassium formiate 14M 78716 100 mL, 500 mL
Potassium phosphate dibasic Solution 3M 79713 100 mL, 500 mL
Potassium phosphate monobasic Solution 1.5M 74056 100 mL, 500 mL
Potassium phosphate monobasic Solution 4M 96183 100 mL, 500 mL
Potassium sodium tartrate Solution 1.5M 81028 100 mL, 500 mL
Potassium sulfate Solution 0.5M 73091 100 mL, 500 mL
Potassium thiocyanate Solution 8M 73059 100 mL, 500 mL
Sodium formiate 7M 74293 100 mL, 500 mL
Sodium phosphate dibasic Solution 0.5M 94046 100 mL, 500 mL
Sodium phosphate monobasic Solution 5M 74092 100 mL, 500 mL
Sodium sulfate solution 1M 83269 250 mL, 1 L
Sodium tartrate dibasic Solution 1.5M 79299 100 mL, 500 mL
Sodium thiocyanate Solution 1M 70331 100 mL
Sodium thiocyanate Solution 8M 80518 100 mL, 500 mL
Trisodium citrate solution 1 M 83273 250 mL, 1 L
Zinc sulfate solution 2M 83265 250 mL, 1 L
back to top
back to X-Ray Crystallography