|
Bacterial Cell Lysis
CelLytic Express Tablets
|
All the advantages of CelLytic™ Express now in a tablet format. Efficient and non-denaturing in-culture protein extraction.
- Save time by eliminating centrifugation steps
- Preserve biological activity with less sample manipulations
- Includes enzymes for complete lysate clarification
- Compatible with affinity resins and magnetic beads
- Ideal for Production Scale Extraction protocols
For lysis of bacterial cultures and direct affinity purification- Eliminating the need for centrifugation!
| Product # |
Product Name |
Package Size |
| C5491 |
CelLytic™ Express Tablets |
25 ea 100 ea |
Compatible with Affinity Purification
1 2 3 4
 |
Lane 1 - Wide MW Marker (M4038)
Lane 2 - CelLytic Express Lysate
Lane 3 - Column Flow-Through
Lane 4 - Elution
CelLytic Express was used for direct, in-culture lysis of BL21 E. coli cells expressing a Metal Affinity Tag (MAT) protein and cultured in Terrific Broth (T9179). The lysate was then immediately purified using the HIS-Select Nickel Affinity Gel (P6611) by standard methods.
|
back to Bacterial Cell Lysis