Sigma-Aldrich
Life Science > Proteomics > Recombinant Protein Expression > Cell Lysis & Protein Extraction > Mammalian Cell Lysis > RIPA Buffer
Mammalian Cell Lysis
RIPA Buffer
RIPA (Radio-Immunoprecipitation Assay) Buffer enables efficient cell lysis and protein solubilization while avoiding protein degradation and interference with the proteins' immunoreactivity and biological activity. RIPA Buffer also results in low background in immunoprecipitation and molecular pull-down assays. Sigma's RIPA Buffer is a ready-to-use 1X solution and is formulated as follows: 150 mM NaCl, 1.0% IGEPAL® CA-630, 0.5% sodium deoxycholate, 0.1% SDS, and 50 mM Tris, pH 8.0. It is also compatible with EZview™ Red Affinity Gels.
Product #  Product Name Package Size
R0278 RIPA Buffer 50 ml
500 ml
back to Mammalian Cell Lysis