Sigma-Aldrich
Life Science > Proteomics > Recombinant Protein Expression > Cell Lysis & Protein Extraction > Plant Cell Lysis
Cell Lysis & Protein Expression
Plant Cell Lysis
We have plant cell lysis kits that offers a convenient method for efficient plant cell lysis and protein solubilization and for the rapid isolation of nuclei and extraction of functional nuclear proteins from plant leaves. CelLytic™ P is a non-denaturing reagent and maintains protein immunoreactivity and biological activity. CelLytic™ PN Extraction Kit is for the rapid isolation of nuclei and extraction of functional nuclear proteins from plant leaves.
  • CelLytic P
  • CelLytic PN Extraction Kit
back to Cell Lysis & Protein Expression