Sigma-Aldrich
Life Science > Proteomics > Recombinant Protein Expression > Cell Lysis & Protein Extraction > SigmaFAST™ Protease Inhibitor Cocktail Tablet
SigmaFAST™ Protease Inhibitor Cocktail Tablet
Broadest Spectrum Protease Inhibition
Water soluble, convenient EDTA-Free tablet protects against:
  • Serine Protease
  • Cysteine Proteases
  • Aspartic Proteases
  • Metalloproteases
And does not interfere downstream applications, such as HIS-tag purification.
Order Now!
Compatible with Nickel Resin | Comparison to Competitor | Related Products | Ordering Information
At the onset of tissue or cell lysis, the extract is teeming with proteolytic enzymes, which will degrade proteins. Protecting your proteins from enzymatic degradation is paramount during the extraction and purification processes. It is equally important to have isolated proteins that are functional and compatible with subsequent downstream applications. The development of this tablet provides broad specificity for inhibition of serine, cysteine, metalloprotease, and aspartic (acidic) proteases and can be used for general purposes or for more specialized applications. This cocktail is provided in a convenient water-soluble tablet and offers superior protection without the presence of EDTA chelator or organic solvent, DMSO.
Compatible with Nickel Resin
Sigma-Aldrich's EDTA-free tablet does not interfere with downstream applications, such as HIS-tag protein purification.
This was tested by repeatedly recovering a HIS-tagged protein from Sigma's HIS-Select Nickel Resin in the presence of the solubilized EDTA-free tablet. As a control, the same HIS-tagged protein was recovered in the presence of PBS (phosphate buffered saline solution). Repeated (4 times) recovery was the same for both sample sets, as measured by protein concentration.
Compatible with Nickel Resin
back to top
Comparison to Competitor
Protease Inhibition using Fluorescent Protease Inhibition Assay.
Protease Inhibition using Fluorescent Protease Inhibition Assay
SDS-PAGE analysis of enzyme digest showing superior protection.
 SDS-PAGE analysis of enzyme digest showing superior protection
A. Control, BSA only
B. BSA, Pepsin Enzyme
C. BSA, Pepsin, SIGMAFAST Protease Inhibitor Cocktail, EDTA-Free Tablet
D. BSA, Pepsin, Competitor EDTA-free Tablet
SDS-PAGE analysis of enzyme digest showing superior protection
A. Control, BSA only
B. BSA, Pancreatin extract
C. BSA, Pancreatin extract, SIGMAFASTä Protease Inhibitor Cocktail, EDTA-Free Tablet
back to top
Related Products
Protease Inhibitor Cocktails
Protease Inhibitor Panel
Phosphatase Inhibitor Cocktails
HIS-Select Products