Sigma-Aldrich
Life Science > Proteomics > Recombinant Protein Expression > Digestion of Protein Complexes
Proteomics & Protien Expression
Digestion of Protein Complexes
cshlp-small Sigma® has prepared product lists for commonly used protocols from Cold Spring Harbor Protocols.
View the entire protocol on BioSupplyNet.com.
Selected Protocol from "Protein-Protein Interactions", 2nd Edition, E. Golemis, P. Adam.
Direct Enzymatic Digestion of Protein Complexes
Sherry Niessen, Ian McLeod and John R. Yates III
Department of Cell Biology, The Scripps Research Institute, La Jolla, California
ABSTRACT
Protein complexes can be digested directly using a number of different protocols. The methods presented here include a double digest with endoproteinase Lys-C and trypsin, or a triple digest with elastase, subtilisin, and trypsin. Endoproteinase Lys-C cleaves carboxy-terminal to lysine residues, while trypsin cleaves carboxy-terminal to lysine and arginine residues. The triple digest uses the relatively nonspecific proteases elastase and subtilisin to create a large number of overlapping peptides for the analysis of posttranslational modifications (MacCoss et al. 2002b). The double and triple digest protocols begin with the reduction and alkylation of cysteine residues to disrupt disulfide bonds and, therefore, higher-order protein structure.   Protein Protein Interactions
Buy this book, Z719145
Products Available for this Protocol
Protocol Material Description Product #  Product Name Add to Cart
Acetone, cold 65051 Acetone
CaCl2, 1 M 21115 Calcium chloride solution
Formic Acid, 90% F0507 Formic acid
Milli-Q water 39253 Water, LC-MS CHROMASOLV®
Trichloroacetic acid (TCA), 20% T9159 Trichloroacetic acid
Tris (2-carboxyethyl)-phosphine hydrochloride (TCEP), 100 mM C4706 Tris(2-carboxyethyl)phosphine hydrochloride
Iodoacetamide (IAA), 500 mM, prepare fresh I1149 Tris(2-carboxyethyl)phosphine hydrochloride
Tris-HCl (or Trizma base), 100 mM (pH 8.0) T6791 Trizma® base
Urea, 8 M in 100 mM Tris-HCl (pH 8.0) U0631 Urea
Elastase Grade II E0127 Elastase, pancreatic from porcine pancreas
Endoproteinase Lys-C P3428 Endoproteinase Lys-C from Lysobacter enzymogenes
Subtilisin P8038 Proteinase, bacterial
Trypsin, sequencing grade, modified T8658 Trypsin from bovine pancreas, suitable for protein sequencing
back to top
View all available product lists for Cold Spring Harbor Laboratory Protocols
Product Association Disclaimer: The Sigma-Aldrich products listed for this specific protocol were selected either to match or to supplement the products listed within the actual protocol. The products/reagents from Sigma-Aldrich have been qualified for usage, but may not have been validated for this specific application. Please refer to the detailed product description on the usage of specific products of interest.