Sigma-Aldrich
Life Science > Proteomics > Recombinant Protein Expression > Protocols > Membrane-bound Peptide Arrays
Proteomics & Protien Expression
Membrane-bound Peptide Arrays
cshlp-small Sigma® has prepared product lists for commonly used protocols from Cold Spring Harbor Protocols.
View the entire protocol on BioSupplyNet.com.
Selected Protocol from "Protein-Protein Interactions", 2nd Edition, E. Golemis, P. Adam.
Synthesis and Probing of Membrane-bound Peptide Arrays
Ronald Frank
Department of Chemical Biology, GBF (German Research Center for Biotechnology)
38124 Braunschweig, Germany
Stefan Dubel
Institute for Biochemistry and Biotechnology, Technical University of Braunschweig
D-38106 Braunschweig, Germany
  Protein Protein Interactions
Buy this book, Z719145
ABSTRACT
The following protocol describes the synthesis of short linear peptides, or peptide pools, on modified cellulose membranes, and the detection of their protein-binding partners. Peptides are synthesized from their carboxyl termini using Fmoc-amino acid derivatives. After completion of the synthesis and cleavage of all side-chain protecting groups, the peptide array on the membrane is incubated with the potential interaction partners to identify their target sequences.
Products Available for this Protocol
Protocol Material Description Product #  Product Name Add to Cart
Buffers, Solutions, and Reagents      
Bromophenol blue indicator (BPB) B0126 Bromophenol Blue
Acetic acid A9967 Acetic acid
DMF D4551 N,N-Dimethylformamide
1-Methyl-2-pyrrolidinone (NMP) 494496 1-Methyl-2-pyrrolidinone
DIC (N,N'-Diisopropylcarbodiimide) D125407 N,N'-Diisopropylcarbodiimide
Acetic anhydride 539996 Acetic anhydride
Piperidine 571261 Piperidine
Methanol or ethanol, technical grade E7148 Ethanol
Trifluoroacetic acid (TFA, synthesis grade) T6508 Trifluoroacetic acid
Triisobutylsilane (TIBS) 278785 Triisobutylsilane
Water W4502 Water
Dichloro­ methane (DCM) 66738 Dichloromethane
Tris-buffered saline (TBS) T6664 Tris Buffered Saline
NaCl S3014 Sodium chloride
KCl P9541 Potassium chloride
Tris-base T6066 Trizma® base
HCl 26612 Hydrogen chloride
NaOH 72068 Sodium hydroxide solution
Tween-20 P9416 TWEEN® 20
Phosphate-buffered saline (PBS) P5368 Phosphate buffered saline
Na2HPO4•2H2O S9763 Sodium phosphate dibasic
KH2PO4 60353 Potassium phosphate dibasic
Citric acid C2404 Citric acid
Casein-based blocking buffer concentrate C7594 Casein blocking buffer
Sucrose S0389 Sucrose
Methanol M3641 Methanol
Sodium dodecyl sulfate (SDS) L3771 Sodium dodecyl sulfate
Alkaline phosphatase (AP)-conjugated detection antibodies Antibody Explorer Antibodies  
Tris-HCl (pH 7.6) T5941 Trizma® hydrochloride
Glycine 50046 Glycine
Horseradish peroxidase (HRP)-conjugated detection antibodies Antibody Explorer Antibodies  
Urea U5378 Urea
Mercaptoethanol M3148 2-Mercaptoethanol
back to top
View all available product lists for Cold Spring Harbor Laboratory Protocols
Product Association Disclaimer: The Sigma-Aldrich products listed for this specific protocol were selected either to match or to supplement the products listed within the actual protocol. The products/reagents from Sigma-Aldrich have been qualified for usage, but may not have been validated for this specific application. Please refer to the detailed product description on the usage of specific products of interest.