|
Proteomics & Protien Expression
Mutant Kinase ATP Analog Pair
 |
The following Cold Spring Harbor Laboratory Press protocols are brought to you by BioSupplyNet in an exclusive partnership with Sigma-Aldrich. Each month, 2 new protocols will be featured, each with a list of products available from Sigma-Aldrich. |
| View the entire protocol on BioSupplyNet.com.
|
|
Selected Protocol from "Protein-Protein Interactions", 2nd Edition, E. Golemis, P. Adam.
Identification of a Mutant Kinase/ATP Analog Pair
Margret B. Einarson and Elena N. Pugacheva Foxx Chase Cancer Center, Philadelphia, PA 19111
Jason R. Orlinck Brigham & Women’s Hospital, Boston, MA 02115
|
| ABSTRACT |
| Protein Kinase cascades regulate many aspects of cellular biochemistry and physiology. Determining the direct substrates of protein kinases is important in understanding how these signaling enzymes exert their effects on cellular processes. A recent development in this area takes advantage of the similarity in the ATP-binding domains of protein kinases. Conserved residues in the ATP-binding site contain large side chains that come into close contact with the N6 position of bound ATP. Mutation of one or more of these large residues to alanine or glycine generates a “pocket” in the ATP-binding site that allows the mutant kinase, but not the wild-type kinase or other cellular kinases, to utilize analogs of ATP with bulky substituents synthesized onto the N6 position. Kinase reactions performed with a mutated kinase and radiolabeled ATP analogs in a cellular lysate allow specific labeling of direct substrates of the mutant kinase, which can later be identified by mass spectrometry or other means. |
|
Buy this book, Z719145
|
| Products Available for this Protocol |
| Protocol Material Description |
Product # |
Product Name |
Add to Cart |
| Buffers & Solutions |
|
|
|
| 50 mm Tris-base (pH 7.4) |
T6066 |
Trizma® base, cell culture tested, meets EP & USP testing specifications, Biotechnology Performance Certified |
|
| 150 mm NaCl |
S3014 |
Sodium chloride for molecular biology |
|
| 10% glycerol |
G5516 |
Glycerol |
|
| 1% Triton X-100 |
X100 |
Triton® X-100 |
|
| 0.5 mm EDTA |
E6758 |
Ethylenediaminetetraacetic acid |
|
| 0.5 mm EGTA |
E3889 |
Ethylene glycol-bis(2-aminoethylether)-N,N,N',N'-tetraacetic acid |
|
| 50 mm NaF |
S7920 |
Sodium fluoride |
|
| 40 mm β-glycerophosphate |
G1150 |
Glycerol 2-phosphate disodium salt hydrate |
|
| 5 mm tetrasodium pyrophosphate |
S6422 |
Sodium pyrophosphate tetrabasic decahydrate |
|
| 0.1 mm sodium vanadate |
72060 |
Sodium metavanadate |
|
| 10 µg/ml aprotinin |
A3428 |
Aprotinin from bovine lung |
|
| 5 µg/ml leupeptin |
L8511 |
Leupeptin hemisulfate salt |
|
| 2 mm phenylmethylsulfonyl fluoride (PMSF), fresh |
P7626 |
Phenylmethanesulfonyl fluoride |
|
| Acetic acid (1%) |
338826 |
Acetic acid |
|
| 250 mm HEPES (pH 7.4) |
H4034 |
HEPES |
|
| 100 mm magnesium acetate |
63052 |
Magnesium acetate solution |
|
| 10 mm dithiothreitol (DTT) |
D9779 |
DL-Dithiothreitol |
|
| 10 µm ATP |
A6559 |
Adenosine 5'-triphosphate disodium salt solution |
|
| PBS |
P5368 |
Phosphate buffered saline |
|
| 25.6 g of Na2HPO4 - 7H2O |
S3264 |
Sodium phosphate dibasic |
|
| 80 g of NaCl |
S3014 |
Sodium chloride |
|
| 2 g of KCl |
P9541 |
Phenylmethanesulfonyl fluoride |
|
| 2 g of KH2PO4 |
P9791 |
Potassium phosphate monobasic |
|
| Ponceau S solution |
78376 |
Ponceau S |
|
| 2x Laemmli sample buffer |
S3401 |
Sample Buffer, Laemmli 2× Concentrate |
|
| 100 mm Tris (pH 6.8) |
T6066 |
Trizma® base, cell culture tested, meets EP & USP testing specifications, Biotechnology Performance Certified |
|
| 2% SDS |
L4390 |
Sodium dodecyl sulfate |
|
| 20% glycerol |
G5516 |
Glycerol |
|
| 4% β-mercaptoethanol (added fresh) |
M3148 |
2-Mercaptoethanol |
|
| Dulbecco's modified Eagle medium (DMEM) |
Cell Culture |
Dulbecco's modified Eagle medium (DMEM) |
|
| Fetal bovine serum (FBS) |
F2442 |
Fetal Bovine Serum |
|
| Cells & Antibodies |
|
|
|
| COS-1 African green monkey kidney epithelial cells (ATCC) |
88031701 |
COS-1 Cell Line from African green monkey kidney |
|
| Antibodies to the epitope tag on the protein kinase of interest |
Antibody Explorer |
Antibody Explorer |
|
| A phospho-specific antibody to a known substrate of your kinase |
Antibody Explorer |
Antibody Explorer |
|
| Additional Reagents |
|
|
|
| BCA protein assay kit |
BCA1 |
Bicinchoninic Acid Kit for Protein Determination |
|
| Nitrocellulose membrane |
N7892 |
Nitrocellulose membranes for blotting |
|
| Vacuum flask |
Z290483 |
Vacuum filtration assembly flasks |
|
back to top View all available Cold Spring Harbor Laboratory Press Protocols |
| |
| Product Association Disclaimer: The Sigma-Aldrich products listed for this specific protocol were selected either to match or to supplement the products listed within the actual protocol. The products/reagents from Sigma-Aldrich have been qualified for usage, but may not have been validated for this specific application. Please refer to the detailed product description on the usage of specific products of interest. |
| |
|