Life Science > Proteomics > Recombinant Protein Expression > Protocols > Proteins from Inclusion Bodies
Proteomics & Protien Expression
Proteins from Inclusion Bodies
cshlp-small Sigma® has prepared product lists for commonly used protocols from Cold Spring Harbor Protocols.
View the entire protocol on BioSupplyNet.com.
Selected Protocol from "Molecular Cloning: A Laboratory Manual", Third Edition, Joseph Sambrook, David W. Russell.
Purification of Expressed Proteins from Inclusion Bodies
Joseph Sambrook
Peter Maccallum Cancer Institute and The University of Melbourne, Australia
David W. Russell
University of Texas Southwestern Medical Center, Dallas
  m8265
Buy this book, M8265
ABSTRACT
The expression of foreign proteins at high levels in E. coli often results in the formation of inclusion bodies composed of insoluble aggregates of the expressed protein. The inclusion bodies are recovered from bacterial lysates by centrifugation and are washed with Triton X-100 and EDTA to remove as much bacterial protein as possible from the aggregated foreign protein.
To obtain soluble protein, the washed inclusion bodies are dissolved in denaturing agents and the released protein is then refolded by gradual removal of the denaturing reagents by dilution or dialysis. Whereas the procedure given here has been used to solubilize inclusion bodies, each protein may require a slightly different procedure, which must be determined empirically.
Products Available for this Protocol
Protocol Material Description Product #  Product Name Add to Cart
Buffers, Solutions, and Reagents      
Tris-Cl (pH 8.0) T5941 Trizma® hydrochloride
NaCl S3014 Sodium chloride
EDTA E7889 Ethylenediaminetetraacetic acid disodium salt solution
Triton X-100 T8787 Triton® X-100, for molecular biology
Deoxycholic acid, protein grade D4297 Deoxycholic acid
HCl H1758 Hydrochloric acid
Urea U5378 Urea
Phenylmethylsulfonyl fluoride (PMSF), fresh P7626 Phenylmethanesulfonyl fluoride
KH2PO4 60353 Potassium phosphate dibasic
KOH P5958 Potassium hydroxide
Isopropanol I9516 2-Propanol, for molecular biology
Enzymes and Gel Loading Buffers      
DNase I AMPD1 DNase I, Amplication Grade
Lysozyme L6751 Lysozyme from chicken egg white, lyophilized powder
Sodium dodecyl sulfate (SDS) L3771 Sodium dodecyl sulfate
Glycerol G5516 Glycerol for molecular biology
Dithiothreitol (DTT) D9779 DL-Dithiothreitol for molecular biology
Bromophenol blue B5525 Bromophenol Blue sodium salt
back to top
View all available product lists for Cold Spring Harbor Laboratory Protocols
Product Association Disclaimer: The Sigma-Aldrich products listed for this specific protocol were selected either to match or to supplement the products listed within the actual protocol. The products/reagents from Sigma-Aldrich have been qualified for usage, but may not have been validated for this specific application. Please refer to the detailed product description on the usage of specific products of interest.