| Protease Inhibitor Cocktails |
| Product Number |
P2714 |
S8820 |
P1860 |
P8465 |
P8340 |
P8849 |
S8830 |
P8215 |
P9599 |
| |
General Use |
General Use |
Tissue Culture |
Bacterial Cells |
Mammalian Tissues |
Histidine-
Tagged
Proteins |
Histidine-
Tagged
Proteins |
Fungal/Yeast Cells |
Plant Cells |
| |
Lyophilized powder -
Water Soluble |
Tablet -
Water Soluble |
Solution in DMSO |
Lyophilized powder -
Vial of DMSO |
Solution in DMSO |
Solution in DMSO |
Tablet -
Water Soluble |
Solution in DMSO |
Solution in DMSO |
| AEBSF |
 |
| Aprotinin |
|
| Bestatin |
 |
| EDTA |
|
| E-64 |
 |
| Leupeptin |
 |
| Pepstatin A |
 |
| Phosphoramidon |
|
| 1,10-Phenanthroline |
 |
back to top
back to Enzyme Explorer
Mix Your Own Cocktails
Create your own broad-spectrum protease inhibitor cocktails or screen your extracts for proteolytic activity. Panel includes inhibitors for serine, cysteine, calpains and metalloproteinases.
| AEBSF |
25 mg |
| Epsilon-AminoCaproic Acid |
25 g |
| Antipain |
5 mg |
| Aprotinin |
5 mg |
| Benzamidine HCl |
5 G |
| Bestatin |
5 mg |
| Chymostatin |
5 mg |
| E-64 |
5 mg |
| EDTA |
50 g |
| N-Ethylmaleimide |
5 g |
| Leupeptin |
5 mg |
| Pepstatin |
5 mg |
| Phosphoramidon |
5 mg |
| Trypsin Inhibitor |
100 mg |
Priced at a fraction of the cost when purchased separately!
Includes detailed instructions for formulating your own cocktails.
back to top
back to Enzyme Explorer
back to top
back to Enzyme Explorer
|