Sigma-Aldrich
Life Science > Proteomics > Recombinant Protein Expression > Purification & Detection > FLAG® System > ANTI-FLAG® M2 Magnetic Beads
Recombinant Protein Purification & Detection
ANTI-FLAG® M2 Magnetic Beads
ANTI-FLAG® M2 Magnetic Beads ImageThe FLAG Expression System is a proven method to express, purify and detect recombinant fusion proteins. Sigma®, the proven provider of FLAG, now offers a magnetic bead for immunoprecipitation, protein purification, and the study of protein-protein interactions.
The ANTI-FLAG M2 Magnetic Bead is composed of murine derived, anti-FLAG M2 monoclonal antibody attached to superparamagnetic iron impregated 4% agarose beads, with an average diameter of 50 µm. The M2 antibody is capable of binding to fusion proteins containing a FLAG peptide sequence at the N-terminus, Met-N-terminus, or C-terminus locations in mammalian, bacterial, and plant extracts.
The magnetic properties allow for:
  • Very rapid separation
  • Significantly accelerated manipulations, such as repetitive washings
  • Processing of multiple samples performed in plate formats
This leads to:
  • Faster experimentation
  • Better reproducibility
  • More accurate quantitation of the proteins of interest
M8823  |  Binding Capacity  |  Purification of FLAG-tagged Baits  |  Automation Overview  |  FLAG Products & Information
Binding Capacity of the ANTI-FLAG M2 Magnetic Beads
Western blot analysis demonstrates that binding capacity of the ANTI-FLAG magnetic beads when compared to the equivalent agarose conjugate (Product No. A2220).
Courtesy of Drs. Guillaume Adlemant and Jarrod Marto, Blais Proteomics Center, Dana-Farber Cancer Institute.
Binding Capacity of the ANTI-FLAG® M2 Magnetic Beads Image
Purification of Two Different FLAG-tagged Baits
Two different FLAG-tagged baits were purified either from a HeLa-S3 nuclear extract (Nuc. Extr.) or from a HeLa-S3 cytoplasmic fraction (Cyto. Extr.). Both samples were prepared from 10e8 cells. The purifications were performed on the Thermo King Fisher magnetic bead processor. 10% of each FLAG immunoprecepitation elution was loaded on the gel and silver stained.
Courtesy of Drs. Guilaume Adelmant and Jarrod Marto, Blais Proteomics Center, Dana-Farber Cancer Institute.
Purification of Two Different FLAG-tagged Baits Image