|
Product No. M8823
Transfer beads to Binding Plate - NUNC 2000 µl
Step 1: FLAG IP
FLAG magnetic beads (50 ml of a 50% slurry)
Note: To prevent the accumulation of beads at the bottom of wells, can be separated in 24 x 10 minutes steps with bead collection between each step.
Transfer beads to Wash Plate 1 - NUNC 2000 µl
Step 2: Wash 1
Transfer beads to Wash Plate 2 - NUNC 2000 µl
Step 3: Wash 2
Transfer beads to Wash Plate 3 - NUNC 2000 µl
Step 4: Wash 3
Transfer beads to Elution Plate 2 - NUNC 300 µl
Step 5: Elution 1
Transfer beads to Elution Plate 2 - NUNC 300 µl
Step 6: Elution 1
IP and Wash Buffer composition:
Elution Buffer composition: (as above with lower NP40 concentration)
|
|||||||||||
|
||||||||||||