Sigma-Aldrich
Life Science > Proteomics > Recombinant Protein Expression > Purification & Detection > FLAG® System > FLAG Immuno-Precipitation Protocol using Magnetic Agarose Beads
Recombinant Protein Purification & Detection
FLAG Immuno-Precipitation Protocol using Magnetic Agarose Beads
Product No. M8823
Transfer beads to Binding Plate - NUNC 2000 µl
Step 1: FLAG IP
FLAG magnetic beads (50 ml of a 50% slurry)
  • Plate: SamplePlate, NUNC 2 ml deepwell plate.
  • Incubation time: 4 hrs
  • Agitation: Yes
Note: To prevent the accumulation of beads at the bottom of wells, can be separated in 24 x 10 minutes steps with bead collection between each step.
Transfer beads to Wash Plate 1 - NUNC 2000 µl
Step 2: Wash 1
  • Volume: 500 µl
  • Incubation time: 5 min
  • Agitation: Yes
Transfer beads to Wash Plate 2 - NUNC 2000 µl
Step 3: Wash 2
  • Volume: 500 µl
  • Incubation time: 5 min
  • Agitation: Yes
Transfer beads to Wash Plate 3 - NUNC 2000 µl
Step 4: Wash 3
  • Volume: 500 µl
  • Incubation time: 5 min
  • Agitation: Yes
Transfer beads to Elution Plate 2 - NUNC 300 µl
Step 5: Elution 1
  • Volume: 50 µl
  • Incubation time: 30 min
  • Agitation: Yes
Transfer beads to Elution Plate 2 - NUNC 300 µl
Step 6: Elution 1
  • Volume: 50 µl
  • Incubation time: 30 min
  • Agitation: Yes
IP and Wash Buffer composition:
  • 150 mM NaCl
  • 50 mM Tris pH 7.5
  • 1 mM EDTA
  • 0.5% NP40
  • 10% Glycerol
Elution Buffer composition: (as above with lower NP40 concentration)
  • 150 mM NaCl
  • 50 mM Tris pH 7.5
  • 1 mM EDTA
  • 0.05% NP40
  • 10% Glycerol
  • FLAG peptide : 500 µg /ml