Sigma-Aldrich
Life Science > Stem Cell Biology > HyStem™ Cell Culture > Extracellular Matrix
HyStem™ Cell Culture
Extracellular Matrix
  • Brief Summary
  • Extracelluar Matrix Types
  • ECM Composition
Brief summary
The extracellular matrix (ECM) is secreted by cells and surrounds them in tissues. It has long been understood to be the structural support for cells since its characteristics set the characteristics of the tissue (i.e. bone compared to cartilage compared to brain)1. However, instead of simply being a passive, mechanical support for cells, it is in fact an extraordinarily complex scaffold composed of a variety of biologically active molecules that are highly regulated and critical for determining the action and fate of the cells that it surrounds4.
Brief summary
The extracellular matrix (ECM) is secreted by cells and surrounds them in tissues. It has long been understood to be the structural support for cells since its characteristics set the characteristics of the tissue (i.e. bone compared to cartilage compared to brain)1. However, instead of simply being a passive, mechanical support for cells, it is in fact an extraordinarily complex scaffold composed of a variety of biologically active molecules that are highly regulated and critical for determining the action and fate of the cells that it surrounds4.
Extracellur Matrix Types
The ECM has two basic forms:
  1. Basement membrane: ECM between epithelial and stromal layers of cells5
  2. Interstitial matrix: ECM surrounding cells forming a porous 3D lattice5
Basement Membrane:
The basement membrane (BM) is a thin layer of ECM that forms between the epithelia and endothelia. It surrounds muscle, fat and nerve cells. It provides mechanical structure, separates different cell types, and signals for cell differentiation, migration, and survival6. A summary of the proteins in the basement membrane and their characterized cell surface receptors are given in Table 16.
Protein Family Cellular Receptor
Laminin Integrins, dystroglycan, heparan sulfate proteoglycans, sulfatides, HNK-1, Lutheran
Collagen IV Integrins
Nidogen/entactin Integrins
Perlecan Dystroglycan
Agrin Dystroglycan, MuSK/agrin receptor
Collagen XVIII Heparan sulfate proteoglycan
Fibulin Integrins
The BM structure is thought to be set by laminin and collagen IV. Laminin has been shown to self assemble into polygonal lattices in vitro6. Likewise, collagen IV also self assembles in vitro through interactions between the carboxy-terminal NC1 (non-collagenase) domains and some slight interactions between the collagen triple helices6. The other protein components of the BM are thought to be non-covalently immobilized in the matrix formed by laminin and collagen IV6.
A form of the basement membrane is commercially available. It is derived from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor that is rich in ECM proteins. Its major components are laminin, collagen IV, heparan sulfate proteoglycan, and nidogen/entactin. In addition to these proteins, there are a variety of growth factors present including bFGF, EGF, IGF-1, PDGF, NGF and TGF-ß1. This same tumor is the primary source for commercially available mouse laminin.
ECM Composition
The ECM is a complex mixture of proteins and glycosaminoglycans (a class of negatively charged polysaccharides). It is composed of three categories of materials:
  1. Glycosaminoglycans and their proteoglycansthat resist compressive forces
  2. Adhesive glycoproteins(laminin, fibronectin, tenascin, nidogen)
  3. Fibrous proteinsthat provide tensile strength (collagens, elastin)
Glycosaminoglycans (GAGs)
GAGs were originally primarily known for being “space fillers” in the ECM. More recently they have been shown to be active signaling molecules whose roles in a variety of cellular processes (including cytokine production, leukocyte recruitment and inflammatory response) are important for controlling cell fate7.
Glycosaminoglycans (GAGs) are linear polysaccharides composed of two basic saccharides: an amino sugar and an uronic acid 2,3. The amino sugar is typically either N-acetyl-D-glucosamine (D-GlcNAc) or N-acetyl-D-galactosamine (D-GalNAc). The uronic acid is either D-glucuronic acid (D-GlcA) or L-iduronic acid (L-IdoA)7. These basic components are further varied by epimerization, sulfation, and deacetylation. The order of the carbohydrate chain and the other chemical modifications determine their specificity and functionality7.
Hyaluronan is the simplest GAG since it is non-sulfated, doesn’t undergo epimerization, and is composed of an unmodified disaccharide repeat. It is also not typically covalently linked to any proteins. The other GAGs are chondroitin sulfate (CS), dermatan sulfate (DS), keratan sulfate (KS), and heparan sulfate (HS). These four GAGs are typically covalently attached to proteins to form proteoglycans. Chondroitin and heparan sulfate are extensively modified by sulfation 1,2,3.
Proteoglycans
A proteoglycan is composed of a core protein with one or more covalently attached GAGs. They are stored in secretory granules, inserted into the plasma membrane or secreted into the ECM8. Table 2 lists several proteoglycans along with their molecular weight, GAG chains, and tissue localization..
Table 2. Proteoglycans present in the ECM8
  ProteoGlycans Core Protein kDA Number of Chains GAG type Tissue
Small interstitial proteoglycans Decori 36 kDa 1 CS Secreted, connective tissue
Biglyca 38 kDa 1-2 CS Secreted, connective tissue
Aggrecan family of matrix proteoglycans Aggrecan 208-220 kDa ~100 CS, HA, KSII Secreted, cartilage
Brevican 96 kDa 0-4 CS Secreted, brain
Neurocan 145 kDa 1-2 CS Secreted, brain
Versican 265 kDa 12-15 CS Secreted, fibroblasts
HS proteoglycans Periecan 400 kDa 1-3 HS Basement membrane
Agrin 212 kDa 2-3 HS Basement membrane
Syndecans 1-4 31-45 kDa 1-2 HS; 1-3 CS HS, CS Epithelial cells, fibroblasts
Betaglycan 110 kDa 1 HS, 1 CD HS Fibroblasts
Glypicans 1-5 ~60 kDa 1-3 HS Epithelial cells, fibroblasts
Serglycin 10-19 kDa 10-15 heparin CS Mast cells
KS proteoglycans Lumican 37 kDa nr KS 1 Broad
Keratocan 37 kDa nr KS 1 Broad
Fibromodulin 59 kDa nr KS 1 Broad
Mimecan 25 kDa nr KS 1 Broad
SV2 80 kDa nr KS 1 Synaptic vesicles
Claustrin 105 kDa nr KS 2 CNS
Glycoproteins
Laminin
Currently there are 15 known heterotrimeric laminins6. Laminin is composed of α, β, and γ chains of which there are 5 α, 4 β, and 6 γ chains2. It has a cross-like structure with 3 short arms and 1 long arm. The α chains possess a large globular domain known as the G domain at the C-termini. This domain is composed of 3 LG domains (LG1-LG3) connected by a linker region to two more LG domains (LG4-LG5). Integrins bind to LG1-3 and dystroglycan to LG4-LG5. Heparan sulfate has been shown to bind to LG4 of the α1 chain6. Laminin is a component of the basement matrix; however, the isoform of laminin present varies with the tissue2. The laminin-1 form is the most prominent in early development and is comprised of α1, β1, and γ1 chains6.
Fibronectin
Fibronectin is a dimer with a molecular weight of ~270 kDa. It exists as a soluble form in blood and body fluids and in fibrils in the ECM. It binds collagen, heparin, other fibronectin proteins, and cell surface integrins. Fibronectin binds integrins through the tri-peptide motif of arginine, glycine, and aspartic acid (RGD)2,3.
Fibrous proteins
Collagen
Collagens are the major structural component of the ECM1. They are the most prevalent protein in the skin and bone, making up 25% of the total protein mass2. Collagens provide scaffolding for the attachment of laminin, proteoglycans and cell surface receptors1. Twenty-eight types of collagens (I–XXVIII) have been identified so far in vertebrates1. Collagens are triple helical proteins that are formed from either homotrimers or heterotrimers of polypeptide chains, referred to as α-chains. α-chains have a three amino acid repeat of Gly-X-Y, where X is typically proline and Y is 4-hydroxyproline (post-translationally modified proline)1,2.
Based on their supramolecular architectures, these types are divided in:
  • fibrillar (I, II, III, V, XI, XXIV and XXVII) – the most common type of collagen, which accounts for 90% of the collagen in the body; it is prominent in bone, skin, tendons, ligaments, and cartilage . Fibrils have a diameter of 10-30 nm in diameter and assembled collagen fibers can have a diameter of 500-3000 nm.
  • fibril-associated (FACIT) (IX, XII, XIV, XVI, XIX, XX, XXI, XXII); these collagens do not form fibrils; instead they attach to fibril-forming collagens; they are thought to order collagen fibrils within the matrix
  • beaded filament (VI)
  • network-forming (IV, VIII and X); these form net-like structures such as in the basement membrane; they also interact with anchoring fibrils (type VII) which link the basement membrane to collagen and laminin in the ECM. Collagen IV self assembles and is instrumental in the formation of the basement membrane.
  • transmembrane collagens (XIII, XVII, XXIII, XXV) These suprastructures have functional roles in cell adhesion, differentiation, tissue development and structural integrity1.
Elastin
Elastin, as its name suggests, provides elasticity to the ECM. It is produced as tropoelastin, a 72 kDa precursor protein and is secreted from the cell. In the extracellular space, it crosslinks with other elastin molecules to form sheets and fibers. Elastin is the primary ECM protein present in arteries where it composes ~50% of their dry weight2.
References
  1. J. Khoshnoodi, J. Cartailler, K. Alvares, A. Veis, B.G. Hudson, “Molecular Recognition in the Assembly of Collagens: Terminal Noncollagenous Domains Are Key Recognition Modules in the Formation of Triple Helical Protomers,” JBC 281(50): 38117–38121 (2006)
  2. J.M. Rhodes, M. Simons, “The extracellular matrix and blood vessel formation: not just a scaffold,” J. Cell. Mol. Med. 11(2): 176-205 (2007).
  3. Essential Cell Biology: An Introduction to the Molecular Biology of the Cell. B. Alberts, D. Bray, A. Johnson, J. Lewis, M. Raff, K. Roberts, P. Walter (editor) Garland Publishing, Inc.
  4. C.Q. Lin, M.J. Bissell, “Multi-faceted regulation of cell differentiation by extracellular matrix”, FASEB Journal 7:737-743 (1993).
  5. Ingber, D. “Mechanical control of tissue morphogenesis during embryological development,” Int. J. Dev. Biol. 50: 255-266 (2006).
  6. T. Sasaki, R. Fassler, E. Hohenester, “Laminin: the crux of basement membrane”, JCB 164(7): 959-963 (2004).
  7. K.R. Taylor, R.L. Gallo, “Glycosaminoglycans and their proteoglycans: host-associated molecular patterns for initiation and modulation of inflammantion,” FASEB Journal 20:9-22 (2006).
  8. Essentials of Glycobiology. A. Varti, R. cummings, J. Esko, H. Freeze, G. Hart, J. Marth (editors) Cold Spring Harbor Laboratory Press (1999)