Sigma-Aldrich
Life Science > Stem Cell Biology > HyStem™ Cell Culture > Why HyStem™? > HyStem™-HP Cell Culture Scaffold Kits
HyStem™ Cell Culture
HyStem™-HP Cell Culture Scaffold Kits
Customizable, well characterized and consistent, HyStem-HP hydrogel cell culture scaffolds provide a complex, three dimensional environment in which cells are able to proliferate, much as they would in‑vivo.
Customizable: Each distinct cell type in a multicellular organism has its own unique microenvironment. HyStem-HP offers the flexibility to tailor the environments of cultured cells to mimic their in vivo counterparts and optimize proliferation and differentiation.
Synthetic: As a synthetic matrix, HyStem-HP offers greater control of the cellular environment than ambiguous extracted ECM alternatives. All components are well characterized and consistently formulated. HyStem-HP contains Hyaluronic acid (synthetic), Extralink (synthetic), Gelin-S (porcine), Heparin and Water.
Biologically accurate: Rich in hyaluronic acid and denatured collagen, the HyStem-HP hydrogel closely mimics the complex three dimensional cellular environments found in life, providing an optimal environment for culturing your cells.
Recommended For...
HyStem-HP is recommended for researchers planning to incorporate and gradually release growth factor into the stem cell environment.
Customize
With HyStem-HP, researchers can tailor the incorporation of ECM proteins and growth factors as well as the elasticity of the hydrogel and cell incorporation (encapsulations or top plating) to create the specific 3D environment required by their cells.
Cell Types
The HyStem Hydrogel Kits are optimal for culturing stem cells whose natural environment is rich in hyaluronic acid (HA).
Cell Encapsulation
The HyStem platform accommodates cell encapsulations (true 3D) as well as plating on top of the hydrogel for pseudo-3D growth.
Cell Attachment
HyStem-C enables the attachment of a wide variety of cell types through Gelin-S (thiolated gelatin).
Growth Factor Incorporation
HyStem-HP enables ionic binding and slow release of growth factors through thiolated heparin; growth factors can be integrated via the matrix rather than cell culture medium. HyStem-HP also enables the attachment of a wide variety of cell types through Gelin-S (thiolated gelatin).
Components
  • HyStem HP (H2541)
  • Gelin-S (G3673)
  • Extralink 2 (E6659)
  • Water, degassed (W3894)
HyStem Cell Kit
Formats:
Trial Kit: (HYSHP010) for 2.5 ml of HyStem-HP Hydrogel
Standard Kit: (HYSHP020) for 7.5 ml of HyStem-HP Hydrogel
Explore:
The HyStem-HP Trial Kit enables researchers to evaluate the HyStem-C technology in their own hands and confirm that HyStem-C is the right choice for their research.
Conditions
HyStem-HP is recommended for both in vivo and in vitro experimentation.
Similar Products
HyStem and HyStem-C
See the HyStem Selection Chart